Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Essential component of the mitotic checkpoint, which prevents cells from prematurely exiting mitosis. Required for the assembly of the dynein-dynactin and MAD1-MAD2 complexes onto kinetochores. Its function related to the spindle assembly machinery is proposed to depend on its association in the mitotic RZZ complex (PubMed:15824131). [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: homolog of Drosophila Zwilch; hZwilch; Protein zwilch homolog; zwilch kinetochore protein; Zwilch, kinetochore associated, homolog
Gene Aliases: 2310031L18Rik; hZwilch; KNTC1AP; ZWILCH
UniProt ID: (Human) Q9H900, (Mouse) Q8R060
Entrez Gene ID: (Human) 55055, (Mouse) 68014, (Rat) 691493
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support