Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence homology: Cow: 92%; Dog: 100%; Horse: 100%; Human: 100%; Rabbit: 77%; Rat: 85%
Probably acts as target recruitment subunit in an E3 ubiquitin ligase complex ZYGA-CUL2-elongin BC.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Protein zyg-11 homolog A; zyg-11 homolog A; ZYG-11A early embryogenesis protein
Gene Aliases: ZYG11; ZYG11A
UniProt ID: (Human) Q6WRX3
Entrez Gene ID: (Human) 440590
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support