Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MEEKYGGDV LAGPGGGGGL GPVDVPSARL TKYIVLLCFT KFLKAVGLFE SYDLLKAVHI VQFIFILKLG TAFFMVLFQK PFSSGKTITK HQIIGSLKIP GRKEFKDKKL NDPRKLVGN (1-118 aa encoded by BC000808 )
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hZTL1; Proton-coupled zinc antiporter SLC30A5; solute carrier family 30 (zinc transporter), member 5; Solute carrier family 30 member 5; Zinc transporter 5; zinc transporter like 1; zinc transporter ZTL1; ZnT-5; ZnT-like transporter 1
Gene Aliases: 1810010K08Rik; AF233321; SLC30A5; UNQ863/PRO1879; ZnT-5; ZNT5; ZNTL1; ZTL1
UniProt ID: (Human) Q8TAD4, (Mouse) Q8R4H9
Entrez Gene ID: (Human) 64924, (Mouse) 69048
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support