Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAPVSWITP GLKTTAVCRG VLRGVTFLLR REGDHEFLEV PEAQEDVEAT FPVHQPGNYS CSYRTDGEGA LSEPSATVTI EELAAPPPPV LMHHGESSQV LHPGNKVTLT CVAPLSGVDF QLRRGEKELL VPRSSTSPDR IFFHLNAVAL GDGGHYTCRY RLHDNQNGWS GDSAPVELIL SDETLPAPEF SPEPESGRAL RLRCLAPLEG ARFALVREDR GGRRVHRFQS PAGTEALFEL HNISVADSAN YSCVYVDLKP PFGGSAPSER LELHVDGPPP RPQLRATWSG AVLAGRDAVL RCEGPIPDVT FELLREGETK AVKTVRTPGA AANLELIFVG PQHAGNYRCR YRSWVPHTFE SELSDPVELL VAES (122-495 aa encoded by B C035719)
Alpha-1-B glycoprotein (A1BG) which is a plasma protein with no known function belongs to the immunoglobulin superfamily. Although the human A1BG has been known for four decades, and the information about the amino acid sequence, chromosomal assignment, and even genetic polymorphism in different populations have been known, no biological function has been suggested. A1BG (Mr approximately equal to 63,000) consists of one polypeptide chain of 474 amino acids with four glucosamine oligosaccharides. The polypeptide has five intrachain disulfide bonds and consists of five repeating structural domains, each containing about 95 amino acids and one disulfide bond. Recently human cysteine-rich secretory protein 3 (CRISP-3) has been found to be a binding partner of A1B, suggesting the A1BG-CRISP-3 complex displays a protecting function from a potentially harmful effect of free CRISP-3 in circulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alpha-1-B glycoprotein; alpha-1-B-glycoprotein; Alpha-1B-glycoprotein; alpha1B-glycoprotein; DKFZp686F0970; epididymis secretory sperm binding protein Li 163pA; HEL-S-163pA
Gene Aliases: A1B; A1BG; ABG; GAB; HYST2477
UniProt ID: (Human) P04217
Entrez Gene ID: (Human) 1
Molecular Function:
immunoglobulin receptor superfamily
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support