Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 1
Additional Information: This antibody is expected to recognize isoform 1 (NP_000671.2) only.
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:16,000.
Adrenergic receptors (ARs) are members of the 7-transmembrane domain G-protein-coupled receptor superfamily that bind the endogenous catecholamines epinephrine and norepinephrine. Pharmacological, structural, and molecular cloning data indicate significant heterogeneity within this receptor family. Nine receptor subtypes have been identified thus far including three alpha-1 AR subtypes (1A/D, 1B, and 1C), three alpha-2 ARs (2A, 2B, and 2C), and three beta AR subtypes (1, 2, and 3). ARs participate in either the onset or maintenance of several disease states including hypertension, cardiac dysfunction (congestive heart failure, ischemia, arrhythmias), diabetes, glaucoma, depression, and impotence. A1AR subtypes are found in numerous tissues and have been shown to be involved in the regulation of blood pressure due to changes in vascular tone and cardiac output. Effects on uterine contraction, hepatic glucose metabolism, heat shock protein 70 (HSP70), proto-oncogene expression, and mitogenesis have been linked to A1AR activation. Norepinephrine can increase nitric oxide synthase (NOS) levels in the hypothalamus by activating alpha-1 AR which may indirectly suppress the release of luteinizing hormone (LH), the androgens, estrogen, and progesterone.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ADA1; adrenergic, alpha-1A-, receptor; Alpha-1A adrenergic receptor; Alpha-1A adrenoceptor; Alpha-1A adrenoreceptor; Alpha-1C adrenergic receptor; Alpha-adrenergic receptor 1c; Alpha1C-AR; G protein coupled receptor
Gene Aliases: ADRA1A; ADRA1C; ADRA1L1; ALPHA1AAR
UniProt ID: (Human) P35348
Entrez Gene ID: (Human) 148
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support