Product References
Curcumol, a major terpenoid from Curcumae Rhizoma, attenuates human uterine leiomyoma cell development via the p38MAPK/NF-κB pathway.
Journal of ethnopharmacology
Yu YH,Zhang HJ,Yang F,Xu L,Liu H
Wed Jun 28 00:00:00 EDT 2023
The protective effect of inhibiting mitochondrial fission on the juvenile rat brain following PTZ kindling through inhibiting the BCL2L13/LC3 mitophagy pathway.
Metabolic brain disease
Fang Q,Zheng S,Chen Q,Chen L,Yang Y,Wang Y,Zhang H,Chen J
Wed Feb 01 00:00:00 EST 2023
HMOX1 Promotes Ferroptosis Induced by Erastin in Lens Epithelial Cell through Modulates Fe2+ Production.
Current eye research
Liao S,Huang M,Liao Y,Yuan C
Sun Jan 01 00:00:00 EST 2023
Pharmacological Activation of Rev-erbα Attenuates Doxorubicin-Induced Cardiotoxicity by PGC-1α Signaling Pathway.
Cardiovascular therapeutics
Zou R,Wang S,Cai H,Wang Y,Wang C
Tue Mar 07 00:00:00 EST 2023
ACTA1 H40Y mutant iPSC-derived skeletal myocytes display mitochondrial defects in an in vitro model of nemaline myopathy.
Experimental cell research
Gartz M,Haberman M,Sutton J,Slick RA,Luttrell SM,Mack DL,Lawlor MW
Wed Mar 15 00:00:00 EDT 2023
ACTA1 H40Y mutant iPSC-derived skeletal myocytes display mitochondrial defects in an in vitro model of nemaline myopathy.
Experimental cell research
Gartz M,Haberman M,Sutton J,Slick RA,Luttrell SM,Mack DL,Lawlor MW
Wed Mar 15 00:00:00 EDT 2023
MFN2 deficiency affects calcium homeostasis in lung adenocarcinoma cells via downregulation of UCP4.
FEBS open bio
Zhang J,Pan L,Zhang Q,Zhao Y,Wang W,Lin N,Zhang S,Wu Q
Thu Jun 01 00:00:00 EDT 2023
Inhibition of TLR4 Alleviates Heat Stroke-Induced Cardiomyocyte Injury by Down-Regulating Inflammation and Ferroptosis.
Molecules (Basel, Switzerland)
Chen D,Geng Y,Deng Z,Li P,Xue S,Xu T,Li G
Wed Mar 01 00:00:00 EST 2023
Gallocatechin Gallate Inhibits the Replication of Pseudorabies Virus via Suppressing the Entry and Release Stages in Its Replication Cycle.
Veterinary sciences
Bo Z,Zhu J,Guo M,Zhang C,Cao Y,Zhang X,Wu Y
Wed Mar 01 00:00:00 EST 2023
Active post-transcriptional regulation and ACLY-mediated acetyl-CoA synthesis as a pivotal target of Shuang-Huang-Sheng-Bai formula for lung adenocarcinoma treatment.
Phytomedicine : international journal of phytotherapy and phytopharmacology
Liu D,Dong C,Wang F,Liu W,Jin X,Qi SL,Liu L,Jin Q,Wang S,Wu J,Wang C,Yang J,Deng H,Cai Y,Yang L,Qin J,Zhang C,Yang X,Wang MS,Yu G,Xue YW,Wang Z,Ge GB,Xu Z,Chen WL
Mon May 01 00:00:00 EDT 2023
Rare sugar L-sorbose exerts antitumor activity by impairing glucose metabolism.
Communications biology
Xu HL,Zhou X,Chen S,Xu S,Li Z,Nakanishi H,Gao XD
Sat Mar 11 00:00:00 EST 2023
miR-146a-5p Modulates Adult Hippocampal Neurogenesis Deficits Through Klf4/p-Stat3 Signaling in APP/PS1 Mice.
Neuroscience
Deng LJ,Wu D,Yang XF,Li T
Mon Aug 21 00:00:00 EDT 2023
The Valsa Mali effector Vm1G-1794 protects the aggregated MdEF-Tu from autophagic degradation to promote infection in apple.
Autophagy
Che R,Liu C,Wang Q,Tu W,Wang P,Li C,Gong X,Mao K,Feng H,Huang L,Li P,Ma F
Thu Jun 01 00:00:00 EDT 2023
Down-regulation of WWP2 aggravates Type 2 diabetes mellitus-induced vascular endothelial injury through modulating ubiquitination and degradation of DDX3X.
Cardiovascular diabetology
You S,Xu J,Yin Z,Wu B,Wang P,Hao M,Cheng C,Liu M,Zhao Y,Jia P,Jiang H,Li D,Cao L,Zhang X,Zhang Y,Sun Y,Zhang N
Sat May 06 00:00:00 EDT 2023
SIRT3 promotes metabolic maturation of human iPSC-derived cardiomyocytes via OPA1-controlled mitochondrial dynamics.
Free radical biology & medicine
Wang R,Xu H,Tan B,Yi Q,Sun Y,Xiang H,Chen T,Liu H,Xie Q,Wang L,Tian J,Zhu J
Wed Feb 01 00:00:00 EST 2023
Lenvatinib inhibits intrahepatic cholangiocarcinoma via Gadd45a-mediated cell cycle arrest.
Discover. Oncology
Yan X,Wang D,Ning Z,Meng ZQ
Thu Feb 23 00:00:00 EST 2023
5'-tiRNA-Gln inhibits hepatocellular carcinoma progression by repressing translation through the interaction with eukaryotic initiation factor 4A-I.
Frontiers of medicine
Wu C,Liu D,Zhang L,Wang J,Ding Y,Sun Z,Wang W
Thu Jun 01 00:00:00 EDT 2023
Aloe-emodin ameliorated MI-induced cardiac remodeling in mice via inhibiting TGF-β/SMAD signaling via up-regulating SMAD7.
Phytomedicine : international journal of phytotherapy and phytopharmacology
Yu J,Zhao X,Yan X,Li W,Liu Y,Wang J,Wang J,Yang Y,Hao Y,Liang Z,Tao Y,Yuan Y,Du Z
Thu Jun 01 00:00:00 EDT 2023
Aloe-emodin ameliorated MI-induced cardiac remodeling in mice via inhibiting TGF-β/SMAD signaling via up-regulating SMAD7.
Phytomedicine : international journal of phytotherapy and phytopharmacology
Yu J,Zhao X,Yan X,Li W,Liu Y,Wang J,Wang J,Yang Y,Hao Y,Liang Z,Tao Y,Yuan Y,Du Z
Thu Jun 01 00:00:00 EDT 2023
Nucleolus assembly impairment leads to two-cell transcriptional repression via NPM1-mediated PRC2 recruitment.
Nature structural & molecular biology
Li D,Cao R,Li Q,Yang Y,Tang A,Zhang J,Liu Q
Sat Jul 01 00:00:00 EDT 2023
G9a inhibition promotes the formation of pacemaker-like cells by reducing the enrichment of H3K9me2 in the HCN4 promoter region.
Molecular medicine reports
Xu P,Jin K,Zhou J,Gu J,Gu X,Dong L,Sun X
Wed Feb 01 00:00:00 EST 2023
Sleep Deprivation Promotes Endothelial Inflammation and Atherogenesis by Reducing Exosomal miR-182-5p.
Arteriosclerosis, thrombosis, and vascular biology
Li X,Cao Y,Xu X,Wang C,Ni Q,Lv X,Yang C,Zhang Z,Qi X,Song G
Thu Jun 01 00:00:00 EDT 2023
Functional investigation of SLC1A2 variants associated with epilepsy.
Cell death & disease
Qu Q,Zhang W,Wang J,Mai D,Ren S,Qu S,Zhang Y
Wed Dec 21 00:00:00 EST 2022
CAMSAP2 promotes colorectal cancer cell migration and invasion through activation of JNK/c-Jun/MMP-1 signaling pathway.
Scientific reports
Wang X,Liu Y,Ding Y,Feng G
Fri Oct 07 00:00:00 EDT 2022
Orexin-A Reverse Bone Mass Loss Induced by Chronic Intermittent Hypoxia Through OX1R-Nrf2/HIF-1α Pathway.
Drug design, development and therapy
Gu H,Ru Y,Wang W,Cai G,Gu L,Ye J,Zhang WB,Wang L
Wed Jul 13 00:00:00 EDT 2022
The RNA polymerase of cytoplasmically replicating Zika virus binds with chromatin DNA in nuclei and regulates host gene transcription.
Proceedings of the National Academy of Sciences of the United States of America
Li P,Wu J,Liu S,Lu R,Jiang H,Wang N,Luo M,Guo L,Xiao J,Bu L,Liu L,Xing F,Peng H,Li C,Ma L,Zhao B,Zhou Z,Guo D
Tue Dec 06 00:00:00 EST 2022
RSL3 triggers glioma stem cell differentiation via the Tgm2/AKT/ID1 signaling axis.
Biochimica et biophysica acta. Molecular basis of disease
Li M,Song D,Chen X,Wang X,Xu L,Yang M,Yang J,Kalvakolanu DV,Wei X,Liu X,Li Y,Guo B,Zhang L
Thu Dec 01 00:00:00 EST 2022
ZC3H4 promotes pulmonary fibrosis via an ER stress-related positive feedback loop.
Toxicology and applied pharmacology
Ghafoor H,Chu H,Huang J,Chen M,Wang S,Wang J,Chao J
Sat Jan 15 00:00:00 EST 2022
ZC3H4 promotes pulmonary fibrosis via an ER stress-related positive feedback loop.
Toxicology and applied pharmacology
Ghafoor H,Chu H,Huang J,Chen M,Wang S,Wang J,Chao J
Sat Jan 15 00:00:00 EST 2022
DJ-1 promotes osteosarcoma progression through activating CDK4/RB/E2F1 signaling pathway.
Frontiers in oncology
Han Z,Wang L,Wang D,Zhang L,Bi Y,Zheng X,Liu W,Bai G,Wang Z,Wan W,Ma Y,Cai X,Liu T,Jia Q
Mon Nov 06 00:00:00 EST 2023
Luteolin alleviates cognitive impairment in Alzheimer's disease mouse model via inhibiting endoplasmic reticulum stress-dependent neuroinflammation.
Acta pharmacologica Sinica
Kou JJ,Shi JZ,He YY,Hao JJ,Zhang HY,Luo DM,Song JK,Yan Y,Xie XM,Du GH,Pang XB
Fri Apr 01 00:00:00 EDT 2022
BUB1B (BUB1 Mitotic Checkpoint Serine/Threonine Kinase B) promotes lung adenocarcinoma by interacting with Zinc Finger Protein ZNF143 and regulating glycolysis.
Bioengineered
Zhou X,Yuan Y,Kuang H,Tang B,Zhang H,Zhang M
Tue Feb 01 00:00:00 EST 2022
Hace1 overexpression mitigates myocardial hypoxia/reoxygenation injury via the effects on Keap1/Nrf2 pathway.
In vitro cellular & developmental biology. Animal
Chen TY,Zheng SK
Sat Oct 01 00:00:00 EDT 2022
Targeting p65 to inhibit Cas3 transcription by Onjisaponin B for radiation damage therapy in p65+/- mice.
Phytomedicine : international journal of phytotherapy and phytopharmacology
Wang TY,Hu YJ,Wang X,Li YF,Zhang F,Yan YD,Dou WT,Cheng CY,Xu P
Thu Sep 01 00:00:00 EDT 2022
LINC00514 facilitates cell proliferation, migration, invasion, and epithelial-mesenchymal transition in non-small cell lung cancer by acting on the Wnt/β-catenin signaling pathway.
Bioengineered
Zhu Y,Wu H,Yang X,Xiong Z,Zhao T,Gan X
Sun May 01 00:00:00 EDT 2022
Erectile dysfunction in hypospadiac male adult rats induced by maternal exposure to di-n-butyl phthalate.
Toxicology
Zhou X,Wang S,Zhou R,Zhang T,Wang Y,Zhang Q,Cong R,Ji C,Luan J,Yao L,Zhou X,Song N
Wed Jun 15 00:00:00 EDT 2022
TXNIP knockout improves cardiac function after myocardial infarction by promoting angiogenesis and reducing cardiomyocyte apoptosis.
Cardiovascular diagnosis and therapy
Wang J,Wang XJ,Zhang Y,Shi WJ,Lei ZD,Jiao XY
Wed Jun 01 00:00:00 EDT 2022
Multi-Omics Analysis of MCM2 as a Promising Biomarker in Pan-Cancer.
Frontiers in cell and developmental biology
Yuan J,Lan H,Huang D,Guo X,Liu C,Liu S,Zhang P,Cheng Y,Xiao S
Sat Jul 16 00:00:00 EDT 2022
Mitochondrial dysfunction by TFAM depletion disrupts self-renewal and lineage differentiation of human PSCs by affecting cell proliferation and YAP response.
Redox biology
Qi Y,Ye Y,Wang R,Yu S,Zhang Y,Lv J,Jin W,Xia S,Jiang W,Li Y,Zhang D
Fri Apr 01 00:00:00 EDT 2022
Scutellarein protects against cardiac hypertrophy via suppressing TRAF2/NF-κB signaling pathway.
Molecular biology reports
Shi X,Hu Y,Jiang Y,Wu J,Zhang C,Zhang J,Wu S,Wu Y,Dong W,Li J
Tue Mar 01 00:00:00 EST 2022
LncRNA BACE1-AS promotes the progression of osteosarcoma through miR-762/SOX7 axis.
Molecular biology reports
Wang C,Zhang T,Yang L,Na X,Qu Y
Fri Jul 01 00:00:00 EDT 2022
Chlorine disinfection elevates the toxicity of polystyrene microplastics to human cells by inducing mitochondria-dependent apoptosis.
Journal of hazardous materials
Qin J,Xia PF,Yuan XZ,Wang SG
Sat Mar 05 00:00:00 EST 2022
A novel SIRT6 activator ameliorates neuroinflammation and ischemic brain injury via EZH2/FOXC1 axis.
Acta pharmaceutica Sinica. B
He T,Shang J,Gao C,Guan X,Chen Y,Zhu L,Zhang L,Zhang C,Zhang J,Pang T
Mon Mar 01 00:00:00 EST 2021
regulates pathological cardiac hypertrophy via a β-catenin-dependent mechanism.
American journal of physiology. Heart and circulatory physiology
Lin L,Xu W,Li Y,Zhu P,Yuan W,Liu M,Shi Y,Chen Y,Liang J,Chen J,Yang B,Cai W,Wen Y,Zhu X,Peng X,Zhou Z,Mo X,Wan Y,Yuan H,Li F,Ye X,Jiang Z,Wang Y,Zhuang J,Fan X,Wu X
Thu Apr 01 00:00:00 EDT 2021
JNK signaling-dependent regulation of histone acetylation are involved in anacardic acid alleviates cardiomyocyte hypertrophy induced by phenylephrine.
PloS one
Peng B,Peng C,Luo X,Wu S,Mao Q,Zhang H,Han X
Tue Jan 25 00:00:00 EST 2022
Overexpression of retinoid X receptor beta provides protection against oxidized low-density lipoprotein-induced inflammation via regulating PGC1α-dependent mitochondrial homeostasis in endothelial cells.
Biochemical pharmacology
Zeng Y,Yan Wang C,Xu J,Le Xu X
Tue Jun 01 00:00:00 EDT 2021
PGC1α Promotes Cisplatin Resistance in Ovarian Cancer by Regulating the HSP70/HK2/VDAC1 Signaling Pathway.
International journal of molecular sciences
Li Y,Kang J,Fu J,Luo H,Liu Y,Li Y,Sun L
Wed Mar 03 00:00:00 EST 2021
Interactions between the ERK1/2 signaling pathway and PCAF play a key role in PE‑induced cardiomyocyte hypertrophy.
Molecular medicine reports
Mao Q,Wu S,Peng C,Peng B,Luo X,Huang L,Zhang H
Wed Sep 01 00:00:00 EDT 2021
Different roles of BAG3 in cardiac physiological hypertrophy and pathological remodeling.
Translational research : the journal of laboratory and clinical medicine
Jia P,Wu N,Yang H,Guo Y,Guo X,Sun Y
Thu Jul 01 00:00:00 EDT 2021
Betulinic acid attenuated bleomycin-induced pulmonary fibrosis by effectively intervening Wnt/β-catenin signaling.
Phytomedicine : international journal of phytotherapy and phytopharmacology
Li X,Liu X,Deng R,Gao S,Jiang Q,Liu R,Li H,Miao Y,Zhai Y,Zhang S,Wang Z,Ren Y,Ning W,Zhou H,Yang C
Fri Jan 01 00:00:00 EST 2021
NAP1L5 Promotes Nucleolar Hypertrophy and Is Required for Translation Activation During Cardiomyocyte Hypertrophy.
Frontiers in cardiovascular medicine
Guo N,Zheng D,Sun J,Lv J,Wang S,Fang Y,Zhao Z,Zeng S,Guo Q,Tong J,Wang Z
Tue Jan 04 00:00:00 EST 2022
Design, synthesis and biological evaluation of novel 1,3,4,9-tetrahydropyrano[3,4-b]indoles as potential treatment of triple negative breast cancer by suppressing PI3K/AKT/mTOR pathway.
Bioorganic & medicinal chemistry
Qin J,Sun X,Ma Y,Cheng Y,Ma Q,Jing W,Qu S,Liu L
Fri Dec 31 00:00:00 EST 2021
Diosgenin exerts anti-tumor effects through inactivation of cAMP/PKA/CREB signaling pathway in colorectal cancer.
European journal of pharmacology
Li SY,Shang J,Mao XM,Fan R,Li HQ,Li RH,Shen DY
Tue Oct 05 00:00:00 EDT 2021
The roles of dipeptidyl peptidase-4 and its inhibitors in the regulation of airway epithelial-mesenchymal transition.
Experimental lung research
Sun J,Chu S,Lu M,Pan Q,Li D,Zheng S,Ma L
Mon Aug 16 00:00:00 EDT 2021
CFIm25-regulated lncRNA acv3UTR promotes gastric tumorigenesis via miR-590-5p/YAP1 axis.
Oncogene
Liu K,Wang BJ,Han W,Chi CH,Gu C,Wang Y,Fu X,Huang W,Liu Z,Song X
Wed Apr 01 00:00:00 EDT 2020
Ginsenoside Rg3 promotes regression from hepatic fibrosis through reducing inflammation-mediated autophagy signaling pathway.
Cell death & disease
Liu X,Mi X,Wang Z,Zhang M,Hou J,Jiang S,Wang Y,Chen C,Li W
Fri Jun 12 00:00:00 EDT 2020
USP22 promotes proliferation in renal cell carcinoma by stabilizing survivin.
Oncology letters
Lin Y,Zhong H,Sun B,Peng Y,Lu F,Chen M,Zhu M,Huang J
Sun Nov 01 00:00:00 EDT 2020
AMPK decreases ERK1/2 activity and cancer cell sensitivity to nutrition deprivation by mediating a positive feedback loop involving eEF2K.
Oncology letters
Tong S,Zhou T,Meng Y,Xu D,Chen J
Wed Jul 01 00:00:00 EDT 2020
5-Azacytidine-Induced Cardiomyocyte Differentiation of Very Small Embryonic-Like Stem Cells.
Stem cells international
Sun X,Li H,Zhu Y,Xu P,Zuo Q,Li B,Gu X
Mon Sep 28 00:00:00 EDT 2020
5-Azacytidine-Induced Cardiomyocyte Differentiation of Very Small Embryonic-Like Stem Cells.
Stem cells international
Sun X,Li H,Zhu Y,Xu P,Zuo Q,Li B,Gu X
Mon Sep 28 00:00:00 EDT 2020
Fucoxanthin inhibits hepatic oxidative stress, inflammation, and fibrosis in diet-induced nonalcoholic steatohepatitis model mice.
Biochemical and biophysical research communications
Takatani N,Kono Y,Beppu F,Okamatsu-Ogura Y,Yamano Y,Miyashita K,Hosokawa M
Thu Jul 23 00:00:00 EDT 2020
Identification of miR-515-3p and its targets, vimentin and MMP3, as a key regulatory mechanism in esophageal cancer metastasis: functional and clinical significance.
Signal transduction and targeted therapy
Hu HF,Xu WW,Zhang WX,Yan X,Li YJ,Li B,He QY
Fri Nov 27 00:00:00 EST 2020
Evaluation of toxicity of halloysite nanotubes and multi-walled carbon nanotubes to endothelial cells in vitro and blood vessels in vivo.
Nanotoxicology
Wu B,Jiang M,Liu X,Huang C,Gu Z,Cao Y
Thu Oct 01 00:00:00 EDT 2020
Moderate-Intensity Continuous Training Improves FGF21 and KLB Expression in Obese Mice.
Biochemistry. Biokhimiia
Xiong Y,Chen Y,Liu Y,Zhang B
Sat Aug 01 00:00:00 EDT 2020
Prognostic Significance and Related Mechanisms of Hexokinase 1 in Ovarian Cancer.
OncoTargets and therapy
Li Y,Tian H,Luo H,Fu J,Jiao Y,Li Y
Mon Apr 18 00:00:00 EDT 2022
CUDC-907 displays potent antitumor activity against human pancreatic adenocarcinoma in vitro and in vivo through inhibition of HDAC6 to downregulate c-Myc expression.
Acta pharmacologica Sinica
Fu XH,Zhang X,Yang H,Xu XW,Hu ZL,Yan J,Zheng XL,Wei RR,Zhang ZQ,Tang SR,Geng MY,Huang X
Wed May 01 00:00:00 EDT 2019
CUDC-907 displays potent antitumor activity against human pancreatic adenocarcinoma in vitro and in vivo through inhibition of HDAC6 to downregulate c-Myc expression.
Acta pharmacologica Sinica
Fu XH,Zhang X,Yang H,Xu XW,Hu ZL,Yan J,Zheng XL,Wei RR,Zhang ZQ,Tang SR,Geng MY,Huang X
Wed May 01 00:00:00 EDT 2019
Therapeutic Effects of Human Urine-Derived Stem Cells in a Rat Model of Cisplatin-Induced Acute Kidney Injury In Vivo and In Vitro.
Stem cells international
Sun B,Luo X,Yang C,Liu P,Yang Y,Dong X,Yang Z,Xu J,Zhang Y,Li L
Thu May 13 00:00:00 EDT 2021
Function of BRD4 in the pathogenesis of high glucose‑induced cardiac hypertrophy.
Molecular medicine reports
Wang Q,Sun Y,Li T,Liu L,Zhao Y,Li L,Zhang L,Meng Y
Tue Jan 01 00:00:00 EST 2019
Function of BRD4 in the pathogenesis of high glucose‑induced cardiac hypertrophy.
Molecular medicine reports
Wang Q,Sun Y,Li T,Liu L,Zhao Y,Li L,Zhang L,Meng Y
Tue Jan 01 00:00:00 EST 2019
Long noncoding RNA Neat1 modulates myogenesis by recruiting Ezh2.
Cell death & disease
Wang S,Zuo H,Jin J,Lv W,Xu Z,Fan Y,Zhang J,Zuo B
Wed Jun 26 00:00:00 EDT 2019
FAM83D inhibits autophagy and promotes proliferation and invasion of ovarian cancer cells via PI3K/AKT/mTOR pathway.
Acta biochimica et biophysica Sinica
Zhu H,Diao S,Lim V,Hu L,Hu J
Thu May 23 00:00:00 EDT 2019
MicroRNA-92b-3p suppresses angiotensin II-induced cardiomyocyte hypertrophy via targeting HAND2.
Life sciences
Yu XJ,Huang YQ,Shan ZX,Zhu JN,Hu ZQ,Huang L,Feng YQ,Geng QS
Sun Sep 01 00:00:00 EDT 2019
lncRNA TUG1 Promotes Cisplatin Resistance by Regulating CCND2 via Epigenetically Silencing miR-194-5p in Bladder Cancer.
Molecular therapy. Nucleic acids
Yu G,Zhou H,Yao W,Meng L,Lang B
Fri Jun 07 00:00:00 EDT 2019
Epigallocatechin-3-gallate inhibits angiotensin II-induced cardiomyocyte hypertrophy via regulating Hippo signaling pathway in H9c2 rat cardiomyocytes.
Acta biochimica et biophysica Sinica
Ma Y,Hu Y,Wu J,Wen J,Li S,Zhang L,Zhang J,Li Y,Li J
Mon Apr 01 00:00:00 EDT 2019
miR-30a-3p Targets MAD2L1 and Regulates Proliferation of Gastric Cancer Cells.
OncoTargets and therapy
Wang Y,Wang F,He J,Du J,Zhang H,Shi H,Chen Y,Wei Y,Xue W,Yan J,Feng Y,Gao Y,Li D,Han J,Zhang J
Tue Apr 12 00:00:00 EDT 2022
Thioredoxin-interacting protein deficiency ameliorates kidney inflammation and fibrosis in mice with unilateral ureteral obstruction.
Laboratory investigation; a journal of technical methods and pathology
Wu M,Li R,Hou Y,Song S,Han W,Chen N,Du Y,Ren Y,Shi Y
Sat Sep 01 00:00:00 EDT 2018
Thioredoxin-interacting protein deficiency ameliorates kidney inflammation and fibrosis in mice with unilateral ureteral obstruction.
Laboratory investigation; a journal of technical methods and pathology
Wu M,Li R,Hou Y,Song S,Han W,Chen N,Du Y,Ren Y,Shi Y
Sat Sep 01 00:00:00 EDT 2018
Gemin5 plays a role in unassembled-U1 snRNA disposal in SMN-deficient cells.
FEBS letters
Jiang D,Zou X,Zhang C,Chen J,Li Z,Wang Y,Deng Z,Wang L,Chen S
Sun Apr 01 00:00:00 EDT 2018
Dihydroartemisinin inhibits the viability of cervical cancer cells by upregulating caveolin 1 and mitochondrial carrier homolog 2: Involvement of p53 activation and NAD(P)H:quinone oxidoreductase 1 downregulation.
International journal of molecular medicine
Zhang T,Hu Y,Wang T,Cai P
Sat Jul 01 00:00:00 EDT 2017
Dihydroartemisinin inhibits the viability of cervical cancer cells by upregulating caveolin 1 and mitochondrial carrier homolog 2: Involvement of p53 activation and NAD(P)H:quinone oxidoreductase 1 downregulation.
International journal of molecular medicine
Zhang T,Hu Y,Wang T,Cai P
Sat Jul 01 00:00:00 EDT 2017
L-PGDS Mediates Vagus Nerve Stimulation-Induced Neuroprotection in a Rat Model of Ischemic Stroke by Suppressing the Apoptotic Response.
Neurochemical research
Zhang L,Ma J,Jin X,Jia G,Jiang Y,Li C
Wed Feb 01 00:00:00 EST 2017
Overexpression of TRIB3 promotes angiogenesis in human gastric cancer.
Oncology reports
Dong S,Xia J,Wang H,Sun L,Wu Z,Bin J,Liao Y,Li N,Liao W
Sat Oct 01 00:00:00 EDT 2016
Overexpression of TRIB3 promotes angiogenesis in human gastric cancer.
Oncology reports
Dong S,Xia J,Wang H,Sun L,Wu Z,Bin J,Liao Y,Li N,Liao W
Sat Oct 01 00:00:00 EDT 2016
Loss of Mouse P2Y6 Nucleotide Receptor Is Associated with Physiological Macrocardia and Amplified Pathological Cardiac Hypertrophy.
The Journal of biological chemistry
Clouet S,Di Pietrantonio L,Daskalopoulos EP,Esfahani H,Horckmans M,Vanorlé M,Lemaire A,Balligand JL,Beauloye C,Boeynaems JM,Communi D
Fri Jul 22 00:00:00 EDT 2016
Loss of Mouse P2Y6 Nucleotide Receptor Is Associated with Physiological Macrocardia and Amplified Pathological Cardiac Hypertrophy.
The Journal of biological chemistry
Clouet S,Di Pietrantonio L,Daskalopoulos EP,Esfahani H,Horckmans M,Vanorlé M,Lemaire A,Balligand JL,Beauloye C,Boeynaems JM,Communi D
Fri Jul 22 00:00:00 EDT 2016
Actin-induced dimerization of palladin promotes actin-bundling.
Protein science : a publication of the Protein Society
Vattepu R,Yadav R,Beck MR
Thu Jan 01 00:00:00 EST 2015
Actin-induced dimerization of palladin promotes actin-bundling.
Protein science : a publication of the Protein Society
Vattepu R,Yadav R,Beck MR
Thu Jan 01 00:00:00 EST 2015
Vinexin β Interacts with Hepatitis C Virus NS5A, Modulating Its Hyperphosphorylation To Regulate Viral Propagation.
Journal of virology
Xiong W,Yang J,Wang M,Wang H,Rao Z,Zhong C,Xin X,Mo L,Yu S,Shen C,Zheng C
Wed Jul 01 00:00:00 EDT 2015
DEAD-box RNA helicase DDX3X inhibits DENV replication via regulating type one interferon pathway.
Biochemical and biophysical research communications
Li G,Feng T,Pan W,Shi X,Dai J
Fri Jan 02 00:00:00 EST 2015
Oncogenic miR-17/20a Forms a Positive Feed-forward Loop with the p53 Kinase DAPK3 to Promote Tumorigenesis.
The Journal of biological chemistry
Cai Z,Cao R,Zhang K,Xue Y,Zhang C,Zhou Y,Zhou J,Sun H,Fu XD
Fri Aug 07 00:00:00 EDT 2015
Transgenic mice overexpressing glia maturation factor-β, an oxidative stress inducible gene, show premature aging due to Zmpste24 down-regulation.
Aging
Imai R,Asai K,Hanai J,Takenaka M
Wed Jul 01 00:00:00 EDT 2015
Transgenic mice overexpressing glia maturation factor-β, an oxidative stress inducible gene, show premature aging due to Zmpste24 down-regulation.
Aging
Imai R,Asai K,Hanai J,Takenaka M
Wed Jul 01 00:00:00 EDT 2015