Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Positive test controls include: Mouse skeletal muscle, Rat skeletal muscle.
Immunogen sequence: DRLEGDHQLL QESLVFDNKH TVYSMEHIRV GWEQLLTSIA RTINEVENQV LTRDAKGLSQ EQLNEFRASF NHFDRKQNGM MEPDDFRACL ISMGYDLGEV E
Alpha-actinin-3, also known as alpha-actinin skeletal muscle isoform 3 or F-actin cross-linking protein, is a protein that in humans is encoded by the ACTN3 gene. This gene encodes a member of the alpha-actin binding protein gene family. The encoded protein is primarily expressed in skeletal muscle and functions as a structural component of sarcomeric Z line. This protein is involved in crosslinking actin containing thin filaments. An allelic polymorphism in this gene results in both coding and non-coding variants; the reference genome represents the coding allele.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Actinin alpha3; actinin, alpha 3; Actinin-alpha3; alpha-actinin skeletal muscle; Alpha-actinin skeletal muscle isoform 3; Alpha-actinin-3; alpha-Actinin3; alphaActinin-3; F-actin cross-linking protein
Gene Aliases: ACTN3
UniProt ID: (Human) Q08043, (Mouse) O88990
Entrez Gene ID: (Human) 89, (Mouse) 11474, (Rat) 171009
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support