Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Knockout to ensure that the antibody binds to the antigen stated.
FIGURE: 1 / 10
Recommended positive controls: A431, HeLa, HepG2, mouse brain, Rat2.
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Alpha catenin is a member of the b-catenin family of cytosolic proteins. Alpha catenin interacts with N-cadherin and b-catenin on the cytosolic side of adherins junctions to mediate the interaction with the actin cytoskeleton. Alpha catenin is a homodimer in solution, but dissociates to form a 1:1 heterodimer with b-catenin in order to interact with cadherins. It also regulates morphological plasticity of synapses and cerebellar and hippocampal lamination during development. Alpha catenin also functions in the control of the startle modulation.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 102 kDa cadherin-associated protein; alpha E catenin; Alpha E-catenin; Alpha N-catenin; alpha(E)-catenin; alpha(N)-catenin; alpha-catenin; Alpha-catenin-related protein; alpha-E-catenin; alpha-N-catenin; alphaE-catenin; alphaN-catenin; cadherin associated protein; Cadherin-associated protein; cadherin-associated protein, related; cancer/testis antigen 114; CAP102; catenin; catenin (cadherin associated protein), alpha 1; catenin (cadherin-associated protein), alpha 1, 102kDa; catenin (cadherin-associated protein), alpha 2; catenin alpha 1; catenin alpha 2; Catenin alpha-1; Catenin alpha-2; Catenin alpha1; Catenin alpha2; catenin, alpha 1; Catenin-alpha1; Catenin-alpha2; cerebellar deficient folia; FLJ36832; FLJ52416; HGNC:2509; NY-REN-13 antigen; Renal carcinoma antigen NY-REN-13
Gene Aliases: 2010010M04Rik; AA517462; AI481747; AI988031; CAP-R; CAP102; CAPR; Catna; Catna1; Catna2; cdf; chp; CT114; CTNNA1; CTNNA2; CTNR; MDPT2
UniProt ID: (Human) P35221, (Human) P26232, (Mouse) P26231, (Mouse) Q61301
Entrez Gene ID: (Human) 1495, (Human) 1496, (Rat) 307505, (Mouse) 12385, (Mouse) 12386, (Rat) 297357
Molecular Function:
non-motor actin binding protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support