Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 6
Description: This L363 monoclonal antibody reacts with alpha-galactosylceramide bound to the mouse MHC class I-like protein CD1d. This complex is recognized by invariant natural killer T (iNKT) cells, a subset of T lymphocytes that expresses the invariant TCR alpha chain Valpha14Jalpha18. Activation of iNKT cells leads to production of Th1 and Th2-associated cytokines as well as dendritic cell maturation.
The L363 antibody has been reported to neutralize iNKT cell activation by alpha-galactosylceramide:CD1d complexes.
Applications Reported: This L363 antibody has been reported for use in functional assays.
Applications Tested: This L363 antibody has been tested by flow cytometric analysis of alpha-galactosylcermide-loaded mouse CD1d-transfected cells. This can be used at less than or equal to 0.25 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
Storage and handling: Use in a sterile environment.
Filtration: 0.2 µm post-manufacturing filtered.
Purity: Greater than 90%, as determined by SDS-PAGE.
Endotoxin Level: Less than 0.001 ng/µg antibody, as determined by LAL assay.
Aggregation: Less than 10%, as determined by HPLC.
This complex is recognized by invariant natural killer T (iNKT) cells, a subset of T lymphocytes that expresses the invariant TCR alpha chain Va14Ja18. Activation of iNKT cells leads to production of Th1 and Th2-associated cytokines as well as dendritic cell maturation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3'-phosphoadenosine-5'-phosphosulfate:GalCer sulfotransferase; 3'-phosphoadenylylsulfate-galactosylceramide 3'-sulfotransferase; 3'-phosphoadenylylsulfate:galactosylceramide 3'-sulfotransferase; Antigen-presenting glycoprotein CD1d1; CD1d.1; Cerebroside sulfotransferase; Galactosylceramide sulfotransferase; GalCer sulfotransferase; T-cell surface glycoprotein CD1d1
Gene Aliases: AI747460; CD1.1; Cd1a; Cd1d; Cd1d1; Cst; Gal3st1; Gcst; Ly-38
UniProt ID: (Mouse) P11609, (Mouse) Q9JHE4
Entrez Gene ID: (Mouse) 12479, (Mouse) 53897
Molecular Function: major histocompatibility complex protein transferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support