Product References
Effect of Sublethal Concentrations of Metal Nanomaterials on Cell Energy Metabolism.
Toxics
Liang C,Jiang Q,Liu Z,Yang J,Zhang J,Zhang S,Xin W
Thu May 11 00:00:00 EDT 2023
Senescent immune cells accumulation promotes brown adipose tissue dysfunction during aging.
Nature communications
Feng X,Wang L,Zhou R,Zhou R,Chen L,Peng H,Huang Y,Guo Q,Luo X,Zhou H
Fri Jun 02 00:00:00 EDT 2023
Small ankyrin 1 (sANK1) promotes docetaxel resistance in castration-resistant prostate cancer cells by enhancing oxidative phosphorylation.
FEBS open bio
Yang Y,Qin H,Ding M,Ji C,Chen W,Diao W,Yin H,Chen M,Gan W,Guo H
Wed Feb 01 00:00:00 EST 2023
VEGFR-3 signaling restrains the neuron-macrophage crosstalk during neurotropic viral infection.
Cell reports
Qi L,Li X,Zhang F,Zhu X,Zhao Q,Yang D,Hao S,Li T,Li X,Tian T,Feng J,Sun X,Wang X,Gao S,Wang H,Ye J,Cao S,He Y,Wang H,Wei B
Tue May 30 00:00:00 EDT 2023
Myeloid-derived suppressor cells ameliorate liver mitochondrial damage to protect against autoimmune hepatitis by releasing small extracellular vesicles.
International immunopharmacology
Shen M,Fan X,Shen Y,Wang X,Wu R,Wang Y,Huang C,Zhao S,Zheng Y,Men R,Luo X,Yang L
Sun Jan 01 00:00:00 EST 2023
Progesterone prevents HGSOC by promoting precancerous cell pyroptosis via inducing fibroblast paracrine.
iScience
Wu N,Zhang X,Wang Z,Zhang X,Fang C,Li H,Zhu M,Wang Y,Liao Q,Chu TY,Wang J
Fri Apr 21 00:00:00 EDT 2023
Niche stiffness sustains cancer stemness via TAZ and NANOG phase separation.
Nature communications
Liu X,Ye Y,Zhu L,Xiao X,Zhou B,Gu Y,Si H,Liang H,Liu M,Li J,Jiang Q,Li J,Yu S,Ma R,Su S,Liao JY,Zhao Q
Mon Jan 16 00:00:00 EST 2023
Omarigliptin Protects the Integrity of the Blood-Brain Barrier After Intracerebral Hemorrhage in Mice.
Journal of inflammation research
Zhang Y,Liu Y,Zhang X,Yong VW,Xue M
Sat Jul 01 00:00:00 EDT 2023
Accumulated cholesterol protects tumours from elevated lipid peroxidation in the microenvironment.
Redox biology
Zhao X,Lian X,Xie J,Liu G
Thu Jun 01 00:00:00 EDT 2023
Pan-HDAC inhibitors augment IL2-induced proliferation of NK cells via the JAK2-STAT5B signaling pathway.
International immunopharmacology
Zheng J,Lu Y,Xiao J,Duan Y,Zong S,Chen X,Hu T,Li L,Zhang Y
Wed Mar 01 00:00:00 EST 2023
System analysis based on the migration- and invasion-related gene sets identifies the infiltration-related genes of glioma.
Frontiers in oncology
Shi S,Zhong J,Peng W,Yin H,Zhong D,Cui H,Sun X
Wed Nov 01 00:00:00 EDT 2023
LCRMP-1 is required for spermatogenesis and stabilises spermatid F-actin organization via the PI3K-Akt pathway.
Communications biology
Chang JH,Chou CH,Wu JC,Liao KM,Luo WJ,Hsu WL,Chen XR,Yu SL,Pan SH,Yang PC,Su KY
Mon Apr 10 00:00:00 EDT 2023
Rational design of a polysaccharide-based viral mimicry nanocomplex for potent gene silencing in inflammatory tissues.
Journal of controlled release : official journal of the Controlled Release Society
Gao H,Wang S,Long Q,Cheng R,Lian W,Koivuniemi A,Ma M,Zhang B,Hirvonen J,Deng X,Liu Z,Ye X,Santos HA
Mon May 01 00:00:00 EDT 2023
SIRT2 promotes cell proliferation and migration through mediating ERK1/2 activation and lactosylceramide accumulation in prostate cancer.
The Prostate
Lin R,Yang Y,Wu E,Zhou M,Wang S,Zhang Q
Sun Jan 01 00:00:00 EST 2023
NETs promote ROS production to induce human amniotic epithelial cell apoptosis via ERK1/2 signaling in spontaneous preterm birth.
American journal of reproductive immunology (New York, N.Y. : 1989)
Hu M,Li H,Li G,Wang Y,Liu J,Zhang M,Shen D,Wang X
Wed Mar 01 00:00:00 EST 2023
Lanthanum Chloride Induces Axon Abnormality Through LKB1-MARK2 and LKB1-STK25-GM130 Signaling Pathways.
Cellular and molecular neurobiology
Song Z,Mao H,Liu J,Sun W,Wu S,Lu X,Jin C,Yang J
Sat Apr 01 00:00:00 EDT 2023
Unusually severe muscular dystrophy upon in-frame deletion of the dystrophin rod domain and lack of compensation by membrane-localized utrophin.
Med (New York, N.Y.)
Gorokhova S,Schessl J,Zou Y,Yang ML,Heydemann PT,Sufit RL,Meilleur K,Donkervoort S,Medne L,Finkel RS,Bönnemann CG
Fri Apr 14 00:00:00 EDT 2023
Arthrocolins Synergizing with Fluconazole Inhibit Fluconazole-Resistant Candida albicans by Increasing Riboflavin Metabolism and Causing Mitochondrial Dysfunction and Autophagy.
Microbiology spectrum
Wu Z,Wu QF,Yuan WL,Chen YH,Hu D,Deng DY,Zhang LL,Niu XM
Mon Feb 27 00:00:00 EST 2023
Splicing factor SNRPA associated with microvascular invasion promotes hepatocellular carcinoma metastasis through activating NOTCH1/Snail pathway and is mediated by circSEC62/miR-625-5p axis.
Environmental toxicology
Mo Z,Li R,Cao C,Li Y,Zheng S,Wu R,Xue J,Hu J,Meng H,Zhai H,Huang W,Zheng F,Zhou B
Mon May 01 00:00:00 EDT 2023
Integrative analysis of the role of BOLA2B in human pan-cancer.
Frontiers in genetics
Liang M,Fei Y,Wang Y,Chen W,Liu Z,Xu D,Shen H,Zhou H,Tang J
Thu Mar 16 00:00:00 EDT 2023
Cardiolipin externalization mediates prion protein (PrP) peptide 106-126-associated mitophagy and mitochondrial dysfunction.
Frontiers in molecular neuroscience
Yang D,Li J,Li Z,Zhao M,Wang D,Sun Z,Wen P,Gou F,Dai Y,Ji Y,Li W,Zhao D,Yang L
Tue Jun 20 00:00:00 EDT 2023
TAT Nanobody Exerts Antiviral Effect against PRRSV In Vitro by Targeting Viral Nucleocapsid Protein.
International journal of molecular sciences
Ren J,Duan H,Dong H,Wu S,Du Y,Zhang G,Zhang A
Wed Jan 18 00:00:00 EST 2023
Site-specific ubiquitination of VDAC1 restricts its oligomerization and mitochondrial DNA release in liver fibrosis.
Experimental & molecular medicine
Wu NN,Wang L,Wang L,Xu X,Lopaschuk GD,Zhang Y,Ren J
Sun Jan 01 00:00:00 EST 2023
Lactate-mediated Fascin protrusions promote cell adhesion and migration in cervical cancer.
Theranostics
Han X,Du S,Chen X,Min X,Dong Z,Wang Y,Zhu C,Wei F,Gao S,Cai Q
Wed May 10 00:00:00 EDT 2023
Side population cells derived from hUCMSCs and hPMSCs could inhibit the malignant behaviors of Tn+ colorectal cancer cells from modifying their O-glycosylation status.
Stem cell research & therapy
Hu W,Ding R,Wang M,Huang P,Wei X,Hu X,Hu T
Fri May 26 00:00:00 EDT 2023
Sperm morphological abnormalities in autosomal dominant polycystic kidney disease are associated with the Hippo signaling pathway via PC1.
Frontiers in endocrinology
Shi WH,Zhou ZY,Ye MJ,Qin NX,Jiang ZR,Zhou XY,Xu NX,Cao XL,Chen SC,Huang HF,Xu CM
Thu May 18 00:00:00 EDT 2023
The role of small GTPases in bisphenol AF-induced multinucleation in comparison with dibutyl phthalate in the male germ cells.
Toxicological sciences : an official journal of the Society of Toxicology
Hu C,Hsiao ZH,Yin L,Yu X
Mon Mar 20 00:00:00 EDT 2023
Integrated proteomic and phosphoproteomic analysis for characterization of colorectal cancer.
Journal of proteomics
Zhu H,Li Y,Guo J,Feng S,Ge H,Gu C,Wang M,Nie R,Li N,Wang Y,Wang H,Zhong J,Qian X,He G
Wed Mar 15 00:00:00 EDT 2023
PPARγ inhibited tumor immune escape by inducing PD-L1 autophagic degradation.
Cancer science
Gou Q,Che S,Chen M,Chen H,Shi J,Hou Y
Sat Jul 01 00:00:00 EDT 2023
Application of deep generative model for design of Pyrrolo[2,3-d] pyrimidine derivatives as new selective TANK binding kinase 1 (TBK1) inhibitors.
European journal of medicinal chemistry
Song S,Tang H,Ran T,Fang F,Tong L,Chen H,Xie H,Lu X
Sun Feb 05 00:00:00 EST 2023
CEBPG suppresses ferroptosis through transcriptional control of SLC7A11 in ovarian cancer.
Journal of translational medicine
Zhang X,Zheng X,Ying X,Xie W,Yin Y,Wang X
Sat May 20 00:00:00 EDT 2023
Bumetanide Rescues Aquaporin-4 Depolarization via Suppressing β-Dystroglycan Cleavage and Provides Neuroprotection in Rat Retinal Ischemia-Reperfusion Injury.
Neuroscience
Chen C,Fan P,Zhang L,Xue K,Hu J,Huang J,Lu W,Xu J,Xu S,Qiu G,Ran J,Gan S
Wed Feb 01 00:00:00 EST 2023
RBX1 regulates PKM alternative splicing to facilitate anaplastic thyroid carcinoma metastasis and aerobic glycolysis by destroying the SMAR1/HDAC6 complex.
Cell & bioscience
Xu D,Yu J,Yang Y,Du Y,Lu H,Zhang S,Feng Q,Yu Y,Hao L,Shao J,Chen L
Tue Feb 21 00:00:00 EST 2023
Effects of the CDC10 (Septin 7) Gene on the Proliferation and Differentiation of Bovine Intramuscular Preadipocyte and 3T3-L1 Cells.
Animals : an open access journal from MDPI
Cheng Z,Li X,Bao S,Yamada T,Cao G,Liu J,Chen A,Tong B
Thu Feb 09 00:00:00 EST 2023
METTL1/WDR4-mediated tRNA m7G modification and mRNA translation control promote oncogenesis and doxorubicin resistance.
Oncogene
Wang Z,Yu P,Zou Y,Ma J,Han H,Wei W,Yang C,Zheng S,Guo S,Wang J,Liu L,Lin S
Thu Jun 01 00:00:00 EDT 2023
Mitigation of non-alcoholic steatohepatitis via recombinant Orosomucoid 2, an acute phase protein modulating the Erk1/2-PPARγ-Cd36 pathway.
Cell reports
Li L,Sun H,Chen J,Ding C,Yang X,Han H,Sun Q
Tue Jul 25 00:00:00 EDT 2023
Geranylgeranylacetone-induced heat shock protein70 expression reduces retinal ischemia-reperfusion injury through PI3K/AKT/mTOR signaling.
Experimental eye research
Zhang L,Xue K,Fan P,Chen C,Hu J,Huang J,Lu W,Xu J,Xu S,Ran J,Zhu S,Gan S
Sat Apr 01 00:00:00 EDT 2023
Daucosterol Alleviates Alcohol-Induced Hepatic Injury and Inflammation through P38/NF-κB/NLRP3 Inflammasome Pathway.
Nutrients
Zhang F,Wang M,Zha Y,Zhou J,Han J,Zhang S
Sun Jan 01 00:00:00 EST 2023
A phosphoglycerate mutase 1 allosteric inhibitor overcomes drug resistance to EGFR-targeted therapy via disrupting IL-6/JAK2/STAT3 signaling pathway in lung adenocarcinoma.
Drug resistance updates : reviews and commentaries in antimicrobial and anticancer chemotherapy
Liang Q,Gong M,Zou JH,Luo MY,Jiang LL,Wang C,Shen NX,Zhang MC,Xu L,Lei HM,Zhang KR,Zhang R,Zhuang G,Zhu L,Chen HZ,Zhou L,Shen Y
Mon May 01 00:00:00 EDT 2023
Dihydrotrichodimerol Purified from the Marine Fungus Acremonium citrinum Prevents NAFLD by Targeting PPARα.
Journal of natural products
Liu J,Gao S,Zhou W,Chen Y,Wang Z,Zeng Z,Zhou H,Lin T
Fri May 26 00:00:00 EDT 2023
Resolvin D1/N-formyl peptide receptor 2 ameliorates paclitaxel-induced neuropathic pain through the activation of IL-10/Nrf2/HO-1 pathway in mice.
Frontiers in immunology
Su CJ,Zhang JT,Zhao FL,Xu DL,Pan J,Liu T
Fri Mar 31 00:00:00 EDT 2023
The TGF-β/Smad2/3 signaling pathway is involved in Musashi2-induced invasion and metastasis of colorectal cancer.
Molecular carcinogenesis
Yu ZL,Liu J,Ning ZK,Tian HK,Wu X,Huang YF,Wu ZC,Zong Z,Zhou TC
Wed Feb 01 00:00:00 EST 2023
RNA-seq reveals Nup62 as a potential regulator for cell division after traumatic brain injury in mice hippocampus.
PeerJ
Zhao J,Wang W,Yan K,Zhao H,Zhang Z,Wang Y,Zhu W,Chen S
Tue Aug 08 00:00:00 EDT 2023
miR-330-3p alleviates the progression of atherosclerosis by downregulating AQP9.
Functional & integrative genomics
Shan E,Yu Y,Tang W,Wang W,Wang X,Zhou S,Gao Y
Tue Mar 07 00:00:00 EST 2023
Luteoloside Induces G0/G1 Phase Arrest of Neuroblastoma Cells by Targeting p38 MAPK.
Molecules (Basel, Switzerland)
He Y,Luo M,Lei S,Zeng Z,Chen T,Wu Y,Wang D,Wang L,Wang L
Sun Feb 12 00:00:00 EST 2023
Recombinant human angiotensin-converting enzyme 2 plays a protective role in mice with sepsis-induced cardiac dysfunction through multiple signaling pathways dependent on converting angiotensin II to angiotensin 1-7.
Annals of translational medicine
Wu C,Chen Y,Zhou P,Hu Z
Sun Jan 15 00:00:00 EST 2023
An integrated in vitro/in silico approach to assess the anti-androgenic potency of isobavachin.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Ren L,Luo H,Zhao J,Huang S,Zhang J,Shao C
Thu Jun 01 00:00:00 EDT 2023
PPARγ agonist pioglitazone enhances colorectal cancer immunotherapy by inducing PD-L1 autophagic degradation.
European journal of pharmacology
Jia X,Qian J,Chen H,Liu Q,Hussain S,Jin J,Shi J,Hou Y
Wed Jul 05 00:00:00 EDT 2023
Enhanced BPGM/2,3-DPG pathway activity suppresses glycolysis in hypoxic astrocytes via FIH-1 and TET2.
Brain research bulletin
E G,Sun B,Liu B,Xu G,He S,Wang Y,Feng L,Wei H,Zhang J,Chen J,Gao Y,Zhang E
Sun Jan 01 00:00:00 EST 2023
DNAJB7 is dispensable for male fertility in mice.
Reproductive biology and endocrinology : RB&E
Bai S,Hu M,Yu L,Chen L,Zhou J,Wu L,Xu B,Jiang X,Zhang X,Tong X,Yue Q
Fri Mar 31 00:00:00 EDT 2023
Circular RNA hsa_circ_0007444 inhibits ovarian cancer progression through miR-23a-3p/DICER1 axis.
Acta biochimica et biophysica Sinica
Zhang M,Sun Y,Xu H,Shi Y,Shen R,Teng F,Xu J,Jia X
Thu Apr 13 00:00:00 EDT 2023
Schisandrin C enhances cGAS-STING pathway activation and inhibits HBV replication.
Journal of ethnopharmacology
Zhao J,Xu G,Hou X,Mu W,Yang H,Shi W,Wen J,Liu T,Wu Z,Bai J,Zhang P,Wang Z,Xiao X,Zou W,Bai Z,Zhan X
Sat Jul 15 00:00:00 EDT 2023
USP22 aggravated diabetic renal tubulointerstitial fibrosis progression through deubiquitinating and stabilizing Snail1.
European journal of pharmacology
Zhao X,He X,Wei W,Huang K
Mon May 15 00:00:00 EDT 2023
Exendin-4 alleviates myocardial ischemia reperfusion injury by enhancing autophagy through promoting nuclear translocation of TFEB.
Experimental cell research
He W,Tong G,Fan H,Zhen C,Zeng L,Xue L,Chen J,Sun Z,He P
Wed Feb 15 00:00:00 EST 2023
DBP induced autophagy and necrotic apoptosis in HepG2 cells via the mitochondrial damage pathway.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Liu H,Xu L,Zhou L,Han W,Li Z,Liu C
Thu Jun 01 00:00:00 EDT 2023
Knockdown of repulsive guidance molecule a promotes polarization of microglia into an anti-inflammatory phenotype after oxygen-glucose deprivation-reoxygenation in vitro.
Neurochemistry international
Shen G,Xiao H,Huang S,Yuan X,Rongrong Z,Ma Y,Qin X
Wed Nov 01 00:00:00 EDT 2023
FTO alleviates cerebral ischemia/reperfusion-induced neuroinflammation by decreasing cGAS mRNA stability in an m6A-dependent manner.
Cellular signalling
Yu Z,Zheng L,Geng Y,Zhang Y,Wang Y,You G,Cai M,Li M,Cheng X,Zan J
Fri Sep 01 00:00:00 EDT 2023
Mice with the Rab10 T73V mutation exhibit anxiety-like behavior and alteration of neuronal functions in the striatum.
Biochimica et biophysica acta. Molecular basis of disease
Zhang J,Li J,You P,Jiang H,Liu Y,Han D,Liu M,Yu H,Su B
Sat Apr 01 00:00:00 EDT 2023
The potential mechanism of qinghua quyu jianpi decoction in the treatment of ulcerative colitis based on network pharmacology and experimental validation.
Journal of ethnopharmacology
Qu F,Li D,Zhang S,Zhang C,Shen A
Wed Jun 28 00:00:00 EDT 2023
Tissue inhibitor of metalloproteinase-4 deletion in mice impacts maternal cardiac function during pregnancy and postpartum.
American journal of physiology. Heart and circulatory physiology
Thurstin AA,Egeli AN,Goldsmith EC,Spinale FG,LaVoie HA
Sun Jan 01 00:00:00 EST 2023
Epileptiform activity influences theta-burst induced LTP in the adult hippocampus: a role for synaptic lipid raft disruption in early metaplasticity?
Frontiers in cellular neuroscience
Carvalho-Rosa JD,Rodrigues NC,Silva-Cruz A,Vaz SH,Cunha-Reis D
Sun May 28 00:00:00 EDT 2023
Nanodrug rescues liver fibrosis via synergistic therapy with H2O2 depletion and Saikosaponin b1 sustained release.
Communications biology
Dong H,Han X,Hao M,Yang Q,Lyu Q,Tang D,Shen Z,Wang K,Kuang H,Cao G,Peng M,Shao M
Thu Feb 16 00:00:00 EST 2023
Ubiquitin-specific protease 24 promotes EV71 infection by restricting K63-linked polyubiquitination of TBK1.
Virologica Sinica
Zang L,Gu J,Yang X,Yuan Y,Guo H,Zhou W,Ma J,Chen Y,Wu Y,Zheng H,Shi W
Wed Feb 01 00:00:00 EST 2023
Chemokine receptor CXCR7 activates Aurora Kinase A and promotes neuroendocrine prostate cancer growth.
The Journal of clinical investigation
Gritsina G,Fong KW,Lu X,Lin Z,Xie W,Agarwal S,Lin D,Schiltz GE,Beltran H,Corey E,Morrissey C,Wang Y,Zhao JC,Hussain M,Yu J
Tue Aug 01 00:00:00 EDT 2023
Hepatitis B virus-related intrahepatic cholangiocarcinoma originates from hepatocytes.
Hepatology international
Song Z,Lin S,Wu X,Ren X,Wu Y,Wen H,Qian B,Lin H,Huang Y,Zhao C,Wang N,Huang Y,Peng B,Li X,Peng H,Shen S
Sun Oct 01 00:00:00 EDT 2023
Ginsenoside Rb1 promotes the activation of PPARα pathway via inhibiting FADD to ameliorate heart failure.
European journal of pharmacology
Li C,Zhang X,Li J,Liang L,Zeng J,Wen M,Pan L,Lv D,Liu M,Cheng Y,Huang H
Mon May 15 00:00:00 EDT 2023
Phosphorylation by IKKβ Promotes the Degradation of HMGCL via NEDD4 in Lung Cancer.
International journal of biological sciences
Zhong C,Xiong G,Yang H,Du X,Du J,Yao F,Fang W,Deng Y
Tue Mar 21 00:00:00 EDT 2023
Strategy To Assess Zoonotic Potential Reveals Low Risk Posed by SARS-Related Coronaviruses from Bat and Pangolin.
mBio
Yang Y,Shen XR,Zhang YL,Jiang RD,Wang X,Guan ZQ,Li Q,Yao YL,Gong QC,Geng R,Wang Q,Zhu Y,Luo JY,Shi ZL,Zhang HL,Peng K,Zhou P
Tue Apr 25 00:00:00 EDT 2023
Magnify is a universal molecular anchoring strategy for expansion microscopy.
Nature biotechnology
Klimas A,Gallagher BR,Wijesekara P,Fekir S,DiBernardo EF,Cheng Z,Stolz DB,Cambi F,Watkins SC,Brody SL,Horani A,Barth AL,Moore CI,Ren X,Zhao Y
Thu Jun 01 00:00:00 EDT 2023
A splicing silencer in SMN2 intron 6 is critical in spinal muscular atrophy.
Human molecular genetics
Wang L,Ji Y,Chen Y,Bai J,Gao P,Feng P
Mon Mar 06 00:00:00 EST 2023
Kaempferol attenuates doxorubicin-induced renal tubular injury by inhibiting ROS/ASK1-mediated activation of the MAPK signaling pathway.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Wu Q,Chen J,Zheng X,Song J,Yin L,Guo H,Chen Q,Liu Y,Ma Q,Zhang H,Yang Q
Sun Jan 01 00:00:00 EST 2023
Role of Type I Cannabinoid Receptor in Sensory Neurons in Psoriasiform Skin Inflammation and Pruritus.
The Journal of investigative dermatology
Liu X,Li L,Jiang J,Ge W,Huang Y,Jin Z,Liu X,Kong Y,Zhanmu O,Zeng X,Li F,Li M,Chen H
Mon May 01 00:00:00 EDT 2023
N6-methyladenosine modification in 18S rRNA promotes tumorigenesis and chemoresistance via HSF4b/HSP90B1/mutant p53 axis.
Cell chemical biology
Chen B,Huang Y,He S,Yu P,Wu L,Peng H
Thu Feb 16 00:00:00 EST 2023
PIM1 attenuates renal ischemia-reperfusion injury by inhibiting ASK1-JNK/P38.
International immunopharmacology
Luo Y,Zhu J,Zhao F,Shi L,Li Y,Wu X
Sun Jan 01 00:00:00 EST 2023
Development of highly potent and specific AKR1C3 inhibitors to restore the chemosensitivity of drug-resistant breast cancer.
European journal of medicinal chemistry
Liu Y,Chen Y,Jiang J,Chu X,Guo Q,Zhao L,Feng F,Liu W,Zhang X,He S,Yang P,Fang P,Sun H
Sun Feb 05 00:00:00 EST 2023
Chronic stress promotes colorectal cancer progression by enhancing glycolysis through β2-AR/CREB1 signal pathway.
International journal of biological sciences
Guan Y,Yao W,Yu H,Feng Y,Zhao Y,Zhan X,Wang Y
Mon May 15 00:00:00 EDT 2023
PIMT is a novel and potent suppressor of endothelial activation.
eLife
Zhang C,Guo ZF,Liu W,Kazama K,Hu L,Sun X,Wang L,Lee H,Lu L,Yang XF,Summer R,Sun J
Tue Apr 18 00:00:00 EDT 2023
Growth differentiation factor 15 is dispensable for acetaminophen-induced liver injury in mice.
Basic & clinical pharmacology & toxicology
Jiang P,Liu Z,Fang T,Zhang Z,Zhang Y,Wang D,Little PJ,Xu S,Weng J
Sat Apr 01 00:00:00 EDT 2023
Supplementation with α-ketoglutarate improved the efficacy of anti-PD1 melanoma treatment through epigenetic modulation of PD-L1.
Cell death & disease
Liu N,Zhang J,Yan M,Chen L,Wu J,Tao Q,Yan B,Chen X,Peng C
Tue Feb 28 00:00:00 EST 2023
Three-in-One Oncolytic Adenovirus System Initiates a Synergetic Photodynamic Immunotherapy in Immune-Suppressive Cholangiocarcinoma.
Small (Weinheim an der Bergstrasse, Germany)
Wang J,Zhu Y,Chen Y,Huang Y,Guo Q,Wang Y,Chen A,Zhou Y,Xu L,Wang L,Zou X,Li X
Tue Aug 01 00:00:00 EDT 2023
PAK2 is essential for chromosome alignment in metaphase I oocytes.
Cell death & disease
Zeng J,Wang S,Gao M,Lu D,Song S,Chen D,Fan W,Xu Z,Zhang Z,Sun X
Wed Feb 22 00:00:00 EST 2023
Enhanced E6AP-mediated ubiquitination of ENO1 via LINC00663 contributes to radiosensitivity of breast cancer by regulating mitochondrial homeostasis.
Cancer letters
Ma J,Zhu J,Li J,Liu J,Kang X,Yu J
Fri Apr 28 00:00:00 EDT 2023
Knockout of TSC2 in Nav1.8+ neurons predisposes to the onset of normal weight obesity.
Molecular metabolism
Brazill JM,Shin D,Magee K,Majumdar A,Shen IR,Cavalli V,Scheller EL
Wed Feb 01 00:00:00 EST 2023
Adipose-specific deletion of the cation channel TRPM7 inhibits TAK1 kinase-dependent inflammation and obesity in male mice.
Nature communications
Zhong W,Ma M,Xie J,Huang C,Li X,Gao M
Mon Jan 30 00:00:00 EST 2023
Differential effects of the LncRNA RNF157-AS1 on epithelial ovarian cancer cells through suppression of DIRAS3- and ULK1-mediated autophagy.
Cell death & disease
Xu P,Xu S,Pan H,Dai C,Xu Y,Wang L,Cong Y,Zhang H,Cao J,Ge L,Jia X
Mon Feb 20 00:00:00 EST 2023
Loss of Lactate/Proton Monocarboxylate Transporter 4 Induces Ferroptosis via the AMPK/ACC Pathway and Inhibition of Autophagy on Human Bladder Cancer 5637 Cell Line.
Journal of oncology
Dong S,Zheng L,Jiang T
Thu Feb 02 00:00:00 EST 2023
MicroMagnify: a multiplexed expansion microscopy method for pathogens and infected tissues.
Research square
Zhao Y,Cheng Z,Stefani C,Skillman T,Klimas A,Lee A,DiBernardo E,Mueller Brown K,Milman T,Gallagher B,Lagree K,Jena B,Pulido J,Filler S,Mitchell A,Hiller L,Lacy-Hulbert A
Mon Mar 06 00:00:00 EST 2023
Novel Self-Assembled Multifunctional Nanoprobes for Second-Near-Infrared-Fluorescence-Image-Guided Breast Cancer Surgery and Enhanced Radiotherapy Efficacy.
Advanced science (Weinheim, Baden-Wurttemberg, Germany)
Zhang YQ,Liu WL,Luo XJ,Shi JP,Zeng YZ,Chen WL,Huang WH,Zhu YY,Gao WL,Li RH,Ming ZH,Zhang LX,Yang RQ,Wang JZ,Zhang GJ
Sat Apr 01 00:00:00 EDT 2023
Down-regulation of WWP2 aggravates Type 2 diabetes mellitus-induced vascular endothelial injury through modulating ubiquitination and degradation of DDX3X.
Cardiovascular diabetology
You S,Xu J,Yin Z,Wu B,Wang P,Hao M,Cheng C,Liu M,Zhao Y,Jia P,Jiang H,Li D,Cao L,Zhang X,Zhang Y,Sun Y,Zhang N
Sat May 06 00:00:00 EDT 2023
Naringenin in Si-Ni-San formula inhibits chronic psychological stress-induced breast cancer growth and metastasis by modulating estrogen metabolism through FXR/EST pathway.
Journal of advanced research
Zhang J,Wang N,Zheng Y,Yang B,Wang S,Wang X,Pan B,Wang Z
Mon May 01 00:00:00 EDT 2023
Polynucleotide phosphorylase protects against renal tubular injury via blocking mt-dsRNA-PKR-eIF2α axis.
Nature communications
Zhu Y,Zhang M,Wang W,Qu S,Liu M,Rong W,Yang W,Liang H,Zeng C,Zhu X,Li L,Liu Z,Zen K
Fri Mar 03 00:00:00 EST 2023
Artesunate delays the dysfunction of age-related intestinal epithelial barrier by mitigating endoplasmic reticulum stress/unfolded protein response.
Mechanisms of ageing and development
Chen H,Sun HM,Wu B,Sun TY,Han LZ,Wang G,Shang YF,Yang S,Zhou DS
Wed Mar 01 00:00:00 EST 2023
Osteogenic microenvironment affects palatal development through glycolysis.
Differentiation; research in biological diversity
Peng X,Chen J,Wang Y,Wang X,Zhao X,Zheng X,Wang Z,Yuan D,Du J
Thu Nov 02 00:00:00 EDT 2023
Maternal NAT10 orchestrates oocyte meiotic cell-cycle progression and maturation in mice.
Nature communications
Jiang X,Cheng Y,Zhu Y,Xu C,Li Q,Xing X,Li W,Zou J,Meng L,Azhar M,Cao Y,Tong X,Qin W,Zhu X,Bao J
Thu Jun 22 00:00:00 EDT 2023
Maternal NAT10 orchestrates oocyte meiotic cell-cycle progression and maturation in mice.
Nature communications
Jiang X,Cheng Y,Zhu Y,Xu C,Li Q,Xing X,Li W,Zou J,Meng L,Azhar M,Cao Y,Tong X,Qin W,Zhu X,Bao J
Thu Jun 22 00:00:00 EDT 2023
CPT1A modulates PI3K/Akt/mTOR pathway to promote preeclampsia.
Placenta
Chen M,Chao B,Xu J,Liu Z,Tao Y,He J,Wang J,Yang H,Luo X,Qi H
Fri Mar 03 00:00:00 EST 2023
Enhanced BCAT1 activity and BCAA metabolism promotes RhoC activity in cancer progression.
Nature metabolism
Qian L,Li N,Lu XC,Xu M,Liu Y,Li K,Zhang Y,Hu K,Qi YT,Yao J,Wu YL,Wen W,Huang S,Chen ZJ,Yin M,Lei QY
Sat Jul 01 00:00:00 EDT 2023
Increased glucose availability sensitizes pancreatic cancer to chemotherapy.
Nature communications
Vaziri-Gohar A,Hue JJ,Abbas A,Graor HJ,Hajihassani O,Zarei M,Titomihelakis G,Feczko J,Rathore M,Chelstowska S,Loftus AW,Wang R,Zarei M,Goudarzi M,Zhang R,Willard B,Zhang L,Kresak A,Willis JE,Wang GM,Tatsuoka C,Salvino JM,Bederman I,Brunengraber H,Lyssiotis CA,Brody JR,Winter JM
Wed Jun 28 00:00:00 EDT 2023
Kaempferol promotes melanogenesis and reduces oxidative stress in PIG1 normal human skin melanocytes.
Journal of cellular and molecular medicine
Xie Y,Mei X,Shi W
Sat Apr 01 00:00:00 EDT 2023
Histone phosphorylation integrates the hepatic glucagon-PKA-CREB gluconeogenesis program in response to fasting.
Molecular cell
Zhao Y,Li S,Chen Y,Wang Y,Wei Y,Zhou T,Zhang Y,Yang Y,Chen L,Liu Y,Hu C,Zhou B,Ding Q
Thu Apr 06 00:00:00 EDT 2023
OTUB1 Catalytic-Independently Deubiquitinates TGFBI and Mediates the Angiogenesis in Infantile Hemangioma by Regulating Glycolysis.
Arteriosclerosis, thrombosis, and vascular biology
Li M,Wang X,Yang E,Li Y,Geng Y,Chen Z,Huang S,Zhang D,Wu H
Mon May 01 00:00:00 EDT 2023
Enamel defects of Axenfeld-Rieger syndrome and the role of PITX2 in its pathogenesis.
Oral diseases
Yang Y,Zhu J,Chiba Y,Fukumoto S,Qin M,Wang X
Wed Nov 01 00:00:00 EDT 2023
Analysis and verification of the circRNA regulatory network RNO_CIRCpedia_ 4214/RNO-miR-667-5p/Msr1 axis as a potential ceRNA promoting macrophage M2-like polarization in spinal cord injury.
BMC genomics
Cao J,Pan C,Zhang J,Chen Q,Li T,He D,Cheng X
Wed Apr 05 00:00:00 EDT 2023
Reduced inhibitory and excitatory input onto parvalbumin interneurons mediated by perineuronal net might contribute to cognitive impairments in a mouse model of sepsis-associated encephalopathy.
Neuropharmacology
Zhang L,Gao YZ,Zhao CJ,Xia JY,Yang JJ,Ji MH
Wed Mar 01 00:00:00 EST 2023
OGG1 aggravates renal ischemia-reperfusion injury by repressing PINK1-mediated mitophagy.
Cell proliferation
Zhao F,Zhu J,Zhang M,Luo Y,Li Y,Shi L,Huang J,Shadekejiang H,Dong S,Wu X
Tue Aug 01 00:00:00 EDT 2023
Tumor-secreted IFI35 promotes proliferation and cytotoxic activity of CD8+ T cells through PI3K/AKT/mTOR signaling pathway in colorectal cancer.
Journal of biomedical science
Li P,Zhou D,Chen D,Cheng Y,Chen Y,Lin Z,Zhang X,Huang Z,Cai J,Huang W,Lin Y,Ke H,Long J,Zou Y,Ye S,Lan P
Wed Jun 28 00:00:00 EDT 2023
Activation of Nrf2 signaling by 4-octyl itaconate attenuates the cartilaginous endplate degeneration by inhibiting E3 ubiquitin ligase ZNF598.
Osteoarthritis and cartilage
Huang B,Wu H,Zheng L,Wei X,Zheng Z,Wu H,Chen J,Shan Z,Liu J,Zhao F
Wed Feb 01 00:00:00 EST 2023
Transportin 1 is a major nuclear import receptor of the nitric oxide synthase interacting protein.
The Journal of biological chemistry
Pörschke M,Rodríguez-González I,Parfentev I,Urlaub H,Kehlenbach RH
Wed Mar 01 00:00:00 EST 2023
Baitouweng decoction alleviates dextran sulfate sodium-induced ulcerative colitis by suppressing leucine-related mTORC1 signaling and reducing oxidative stress.
Journal of ethnopharmacology
Wei P,He Q,Liu T,Zhang J,Shi K,Zhang J,Liu S
Sat Mar 25 00:00:00 EDT 2023
Nanoemulsions of Hydroxysafflor Yellow A for Enhancing Physicochemical and In Vivo Performance.
International journal of molecular sciences
Zhang Y,Zhong C,Wang Q,Zhang J,Zhao H,Huang Y,Zhao D,Yang J
Fri May 12 00:00:00 EDT 2023
Norbergenin prevents LPS-induced inflammatory responses in macrophages through inhibiting NFκB, MAPK and STAT3 activation and blocking metabolic reprogramming.
Frontiers in immunology
Li W,Cai Z,Schindler F,Bahiraii S,Brenner M,Heiss EH,Weckwerth W
Fri Jun 02 00:00:00 EDT 2023
Metrnl Alleviates Lipid Accumulation by Modulating Mitochondrial Homeostasis in Diabetic Nephropathy.
Diabetes
Zhou Y,Liu L,Jin B,Wu Y,Xu L,Chang X,Hu L,Wang G,Huang Y,Song L,Zhang T,Wang Y,Xiao Y,Zhang F,Shi M,Liu L,Wang T,Yan R,Guo B
Mon May 01 00:00:00 EDT 2023
RAB7A GTPase Is Involved in Mitophagosome Formation and Autophagosome-Lysosome Fusion in N2a Cells Treated with the Prion Protein Fragment 106-126.
Molecular neurobiology
Li Z,Lai M,Li J,Yang D,Zhao M,Wang D,Sun Z,Wen P,Gou F,Dai Y,Ji Y,Zhao D,Qiao J,Yang L
Wed Mar 01 00:00:00 EST 2023
Upregulation of LPGAT1 Enhances Lung Adenocarcinoma Proliferation.
Frontiers in bioscience (Landmark edition)
Gong H,Ma C,Li X,Zhang X,Zhang L,Chen P,Wang W,Hu Y,Huang T,Wu N,Wang X
Thu May 11 00:00:00 EDT 2023
Saponin extract from Achyranthes bidentata Blume alleviates disuse-induced muscle atrophy through PI3K/Akt signaling pathway.
Journal of ethnopharmacology
Shi Y,Zhang ZW,Du MM,Wu J,Li JX
Thu Aug 10 00:00:00 EDT 2023
A novel variant in the FBP1 gene causes fructose-1,6-bisphosphatase deficiency through increased ubiquitination.
Archives of biochemistry and biophysics
Liang X,Liu X,Li W,Zhang L,Zhang B,Lai G,Zhao Y
Sat Jul 01 00:00:00 EDT 2023
SARS-CoV-2 Z-RNA activates the ZBP1-RIPK3 pathway to promote virus-induced inflammatory responses.
Cell research
Li S,Zhang Y,Guan Z,Ye M,Li H,You M,Zhou Z,Zhang C,Zhang F,Lu B,Zhou P,Peng K
Wed Mar 01 00:00:00 EST 2023
Macrophages maintain mammary stem cell activity and mammary homeostasis via TNF-α-PI3K-Cdk1/Cyclin B1 axis.
NPJ Regenerative medicine
Zhou Y,Ye Z,Wei W,Zhang M,Huang F,Li J,Cai C
Tue May 02 00:00:00 EDT 2023
MitoQ Protects Ovarian Organoids against Oxidative Stress during Oogenesis and Folliculogenesis In Vitro.
International journal of molecular sciences
Wang J,Du H,Ma L,Feng M,Li L,Zhao X,Dai Y
Wed Jan 04 00:00:00 EST 2023
Sanggenon C inhibits cell proliferation and induces apoptosis by regulating the MIB1/DAPK1 axis in glioblastoma.
MedComm
Chang H,Hou J,Shao Y,Xu M,Weng X,Du Y,Shi J,Zhang L,Cui H
Tue Aug 01 00:00:00 EDT 2023
Rationally designed donepezil-based hydroxamates modulate Sig-1R and HDAC isoforms to exert anti-glioblastoma effects.
European journal of medicinal chemistry
Nepali K,Wu AC,Lo WL,Chopra B,Lai MJ,Chuang JY,Liou JP
Wed Feb 15 00:00:00 EST 2023
5'-tiRNA-Gln inhibits hepatocellular carcinoma progression by repressing translation through the interaction with eukaryotic initiation factor 4A-I.
Frontiers of medicine
Wu C,Liu D,Zhang L,Wang J,Ding Y,Sun Z,Wang W
Thu Jun 01 00:00:00 EDT 2023
Hawthorn leaves flavonoids attenuate cardiac remodeling induced by simulated microgravity.
Pharmaceutical biology
Liu T,Ma Y,Zhao H,Wang P,Niu Y,Hu Y,Shen X,Zhang M,Yan B,Yu J
Fri Dec 01 00:00:00 EST 2023
Efficient correction of ABCA4 variants by CRISPR-Cas9 in hiPSCs derived from Stargardt disease patients.
Molecular therapy. Nucleic acids
Siles L,Ruiz-Nogales S,Navinés-Ferrer A,Méndez-Vendrell P,Pomares E
Tue Jun 13 00:00:00 EDT 2023
ARL9 is upregulated and serves as a biomarker for a poor prognosis in colon adenocarcinoma.
BMC gastroenterology
Yang HD,Jin XX,Gu BB,Li D,Yan LL
Thu Feb 23 00:00:00 EST 2023
Sulforaphane attenuates microglia-mediated neuronal damage by down-regulating the ROS/autophagy/NLRP3 signal axis in fibrillar Aβ-activated microglia.
Brain research
Yang Y,Zhang J,Yang C,Dong B,Fu Y,Wang Y,Gong M,Liu T,Qiu P,Xie W,Lü T
Wed Feb 15 00:00:00 EST 2023
Formin protein DIAPH1 positively regulates PD-L1 expression and predicts the therapeutic response to anti-PD-1/PD-L1 immunotherapy.
Clinical immunology (Orlando, Fla.)
Mei J,Cai Y,Wang H,Xu R,Zhou J,Lu J,Yang X,Pan J,Liu C,Xu J,Zhu Y
Sun Jan 01 00:00:00 EST 2023
Role of NF-κB in lead exposure-induced activation of astrocytes based on bioinformatics analysis of hippocampal proteomics.
Chemico-biological interactions
Fan S,Weixuan W,Han H,Liansheng Z,Gang L,Jierui W,Yanshu Z
Wed Jan 25 00:00:00 EST 2023
CNTN1 in the Nucleus Accumbens is Involved in Methamphetamine-Induced Conditioned Place Preference in Mice.
Neurotoxicity research
Zhang L,Zeng Z,Lu X,Li M,Yao J,Zou G,Chen Z,Li Q,Li C,Li F
Tue Aug 01 00:00:00 EDT 2023
Downregulation of AC092894.1 promotes oxaliplatin resistance in colorectal cancer via the USP3/AR/RASGRP3 axis.
BMC medicine
Zheng Z,Wu M,Li H,Xu W,Yang M,Pan K,Ni Y,Jiang T,Zheng H,Jin X,Zhang Y,Ding L,Fu J
Mon Apr 03 00:00:00 EDT 2023
The Lnc-RNA APPAT Suppresses Human Aortic Smooth Muscle Cell Proliferation and Migration by Interacting With MiR-647 and FGF5 in Atherosclerosis.
Journal of endovascular therapy : an official journal of the International Society of Endovascular Specialists
Meng F,Han L,Liang Q,Lu S,Huang Y,Liu J
Fri Dec 01 00:00:00 EST 2023
Ligation to Scavenging Strategy Enables On-Demand Termination of Targeted Protein Degradation.
Journal of the American Chemical Society
Jin Y,Fan J,Wang R,Wang X,Li N,You Q,Jiang Z
Wed Apr 05 00:00:00 EDT 2023
Allograft inflammatory factor 1 is a potential diagnostic, immunological, and prognostic biomarker in pan-cancer.
Aging
Liu X,Zhang D,Hu J,Xu S,Xu C,Shen Y
Mon Apr 03 00:00:00 EDT 2023
Differences in histomorphology and expression of key lipid regulated genes of four adipose tissues from Tibetan pigs.
PeerJ
Lin C,Dong Z,Song J,Wang S,Yang Y,Li H,Feng Z,Pei Y
Tue Jan 17 00:00:00 EST 2023
Yi-Shen-Xie-Zhuo formula alleviates cisplatin-induced AKI by regulating inflammation and apoptosis via the cGAS/STING pathway.
Journal of ethnopharmacology
Qi J,Luo Q,Zhang Q,Wu M,Zhang L,Qin L,Xue Q,Nie X
Mon Jun 12 00:00:00 EDT 2023
Excessive ER-phagy contributes to ochratoxin A-induced apoptosis.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Deng H,Chen W,Zhang B,Zhang Y,Han L,Zhang Q,Yao S,Wang H,Shen XL
Thu Jun 01 00:00:00 EDT 2023
MIR222HG attenuates macrophage M2 polarization and allergic inflammation in allergic rhinitis by targeting the miR146a-5p/TRAF6/NF-κB axis.
Frontiers in immunology
Wen S,Li F,Tang Y,Dong L,He Y,Deng Y,Tao Z
Fri May 26 00:00:00 EDT 2023
LncRNA SLC25A21-AS1 increases the chemosensitivity and inhibits the progression of ovarian cancer by upregulating the expression of KCNK4.
Functional & integrative genomics
Huang K,Chen X,Geng Z,Xiong X,Cong Y,Pan X,Liu S,Ge L,Xu J,Jia X
Thu Mar 30 00:00:00 EDT 2023
Moshen granule ameliorates membranous nephropathy by blocking intrarenal renin-angiotensin system signalling via the Wnt1/β-catenin pathway.
Phytomedicine : international journal of phytotherapy and phytopharmacology
Wang YN,Miao H,Hua MR,Yang JZ,Pei M,Yu HX,Wei LJ,Zou L,Zhang YM,Cao G,Zhao YY
Thu Jun 01 00:00:00 EDT 2023
Necroptosis of macrophage is a key pathological feature in biliary atresia via GDCA/S1PR2/ZBP1/p-MLKL axis.
Cell death & disease
Yang S,Chang N,Li W,Yang T,Xue R,Liu J,Zhang L,Yao X,Chen Y,Wang H,Yang L,Huang J,Li L
Wed Mar 01 00:00:00 EST 2023
A novel role of Fas in delaying cellular senescence.
Heliyon
Saha C,Li J,Sun X,Liu X,Huang G
Wed Feb 01 00:00:00 EST 2023
Development of a series of quinazoline-2,5-diamine derivatives as potent hematopoietic progenitor kinase 1 (HPK1) inhibitors.
European journal of medicinal chemistry
Shi H,Tang H,Li Y,Chen D,Liu T,Chen Y,Wang X,Chen L,Wang Y,Xie H,Xiong B
Wed Feb 15 00:00:00 EST 2023
Qi Gong Wan ameliorates adipocyte hypertrophy and inflammation in adipose tissue in a PCOS mouse model through the Nrf2/HO-1/Cyp1b1 pathway: Integrating network pharmacology and experimental validation in vivo.
Journal of ethnopharmacology
Zheng R,Shen H,Li J,Zhao J,Lu L,Hu M,Lin Z,Ma H,Tan H,Hu M,Li J
Mon Jan 30 00:00:00 EST 2023
NAMPT inhibition reduces macrophage inflammation through the NAD+/PARP1 pathway to attenuate liver ischemia-reperfusion injury.
Chemico-biological interactions
Lu J,Wang M,Chen Y,Song H,Wen D,Tu J,Guo Y,Liu Z
Thu Jan 05 00:00:00 EST 2023
p38-mediated FOXN3 phosphorylation modulates lung inflammation and injury through the NF-κB signaling pathway.
Nucleic acids research
Zhu X,Huang B,Zhao F,Lian J,He L,Zhang Y,Ji L,Zhang J,Yan X,Zeng T,Ma C,Liang Y,Zhang C,Lin J
Tue Mar 21 00:00:00 EDT 2023
Tumor-intrinsic IRE1α signaling controls protective immunity in lung cancer.
Nature communications
Crowley MJP,Bhinder B,Markowitz GJ,Martin M,Verma A,Sandoval TA,Chae CS,Yomtoubian S,Hu Y,Chopra S,Tavarez DA,Giovanelli P,Gao D,McGraw TE,Altorki NK,Elemento O,Cubillos-Ruiz JR,Mittal V
Mon Jan 09 00:00:00 EST 2023
Targeting plasmodium α-tubulin-1 to block malaria transmission to mosquitoes.
Frontiers in cellular and infection microbiology
Zhang G,Niu G,Hooker-Romera D,Shabani S,Ramelow J,Wang X,Butler NS,James AA,Li J
Tue Apr 04 00:00:00 EDT 2023
Rift Valley Fever Virus Nucleoprotein Triggers Autophagy to Dampen Antiviral Innate Immune Responses.
Journal of virology
Zhu X,Guan Z,Fang Y,Zhang Y,Guan Z,Li S,Peng K
Thu Apr 27 00:00:00 EDT 2023
Cardamonin targets KEAP1/NRF2 signaling for protection against atherosclerosis.
Food & function
Fan P,Meng H,Hao W,Zheng Y,Li H,Zhang Z,Du L,Guo X,Wang D,Wang Y,Wu H
Mon May 22 00:00:00 EDT 2023
Recognition of an Ala-rich C-degron by the E3 ligase Pirh2.
Nature communications
Wang X,Li Y,Yan X,Yang Q,Zhang B,Zhang Y,Yuan X,Jiang C,Chen D,Liu Q,Liu T,Mi W,Yu Y,Dong C
Sat Apr 29 00:00:00 EDT 2023
DR1-CSE/H2S pathway upregulates autophagy and inhibits H9C2 cells damage induced by high glucose.
Acta cardiologica
Li H,Wei Y,Xi Y,Jiao L,Wen X,Wu R,Chang G,Sun F,Hao J
Sat Jul 01 00:00:00 EDT 2023
Bone marrow mesenchymal stem cell-derived exosomes reduce insulin resistance and obesity in mice via the PI3K/AKT signaling pathway.
FEBS open bio
Shi H,Hao X,Sun Y,Zhang H,Zhao Y,Wang B,Lu J,Hou W,Yan Y,Yu X,Xue L,Luo X,Wang H
Thu Jun 01 00:00:00 EDT 2023
Transcriptional coactivation by EHMT2 restricts glucocorticoid-induced insulin resistance in a study with male mice.
Nature communications
Lee RA,Chang M,Yiv N,Tsay A,Tian S,Li D,Poulard C,Stallcup MR,Pufall MA,Wang JC
Tue May 30 00:00:00 EDT 2023
Bioinformatics analysis of ferroptosis-related gene AKR1C3 as a potential biomarker of asthma and its identification in BEAS-2B cells.
Computers in biology and medicine
Wang Y,Fan J,Tong Y,Wang L,Wang L,Weng C,Lai C,Song J,Zhang W
Mon May 01 00:00:00 EDT 2023
Choroid Plexus Modulates Subventricular Zone Adult Neurogenesis and Olfaction Through Secretion of Small Extracellular Vesicles.
bioRxiv : the preprint server for biology
Liu LL,Shannahan J,Zheng W
Sat Mar 18 00:00:00 EDT 2023
N-methylpiperazine-diepoxyovatodiolide ameliorates peritoneal fibrosis via suppressing TGF-β/Smad and JAK/STAT signaling pathway.
Chemico-biological interactions
Mo M,Zeng Y,Zeng Y,Li S,He X,Chen X,Luo Q,Liu M,Luo C,Dou X,Peng F,Long H
Fri Sep 01 00:00:00 EDT 2023
Fisetin treatment alleviates kidney injury in mice with diabetes-exacerbated atherosclerosis through inhibiting CD36/fibrosis pathway.
Acta pharmacologica Sinica
Zou TF,Liu ZG,Cao PC,Zheng SH,Guo WT,Wang TX,Chen YL,Duan YJ,Li QS,Liao CZ,Xie ZL,Han JH,Yang XX
Sun Oct 01 00:00:00 EDT 2023
Inhibition of Oocyte Maturation by Follicular Extracellular Vesicles of Nonhyperandrogenic PCOS Patients Requiring IVF.
The Journal of clinical endocrinology and metabolism
Liu C,Wang M,Yao H,Cui M,Gong X,Wang L,Sui C,Zhang H
Wed May 17 00:00:00 EDT 2023
Deciphering the underlying wound healing mechanisms of Streptocaulon juventas (Lour.) Merr. by integrating network pharmacology, transcriptomics, and experimental validation.
Journal of ethnopharmacology
Li Y,Zang J,Wang X,Feng X,Qiu F
Fri Feb 10 00:00:00 EST 2023
Dual Antioxidant DH-217 Mitigated Cerebral Ischemia-Reperfusion Injury by Targeting IKKβ/Nrf2/HO-1 Signal Axis.
Neurochemical research
Shen M,Zheng Y,Li G,Chen Y,Huang L,Wu J,Hong C
Wed Feb 01 00:00:00 EST 2023
LncRNA DANCR deficiency promotes high glucose-induced endothelial to mesenchymal transition in cardiac microvascular cells via the FoxO1/DDAH1/ADMA signaling pathway.
European journal of pharmacology
Wu M,Li T,Li G,Niu B,Wu T,Yan L,Wang S,He S,Huang C,Tong W,Li N,Jiang J
Wed Jul 05 00:00:00 EDT 2023
MYO10 regulates genome stability and cancer inflammation through mediating mitosis.
Cell reports
Mayca Pozo F,Geng X,Miyagi M,Amin AL,Huang AY,Zhang Y
Tue May 30 00:00:00 EDT 2023
MAD2B Blunts Chronic Unpredictable Stress and Corticosterone Stimulation-Induced Depression-Like Behaviors in Mice.
The international journal of neuropsychopharmacology
Wang XL,Miao C,Su Y,Zhang C,Meng X
Tue Feb 14 00:00:00 EST 2023
Forsythiaside A Improves the Inhibitory Efficiency of Recombinant Protein Vaccines against Bovine Viral Diarrhea Virus Infection.
International journal of molecular sciences
Yang G,Wang J,Wang S,Zhu Y
Sat Aug 20 00:00:00 EDT 2022
Water Treadmill Training Ameliorates Neurite Outgrowth Inhibition Associated with NGR/RhoA/ROCK by Inhibiting Astrocyte Activation following Spinal Cord Injury.
Oxidative medicine and cellular longevity
Ying X,Yu X,Zhu J,Li X,Zheng Y,Xie Q,Wu Q,Li S,Yue J,Zhou Y,Zhou K,Tu W,Jiang S
Fri Apr 08 00:00:00 EDT 2022
The m6A demethylase ALKBH5 promotes tumor progression by inhibiting RIG-I expression and interferon alpha production through the IKKε/TBK1/IRF3 pathway in head and neck squamous cell carcinoma.
Molecular cancer
Jin S,Li M,Chang H,Wang R,Zhang Z,Zhang J,He Y,Ma H
Sat Apr 09 00:00:00 EDT 2022
Opposing reduced VPAC1 and enhanced VPAC2 VIP receptors in the hippocampus of the Li2+-pilocarpine rat model of temporal lobe epilepsy.
Neurochemistry international
Serpa A,Bento M,Caulino-Rocha A,Pawlak S,Cunha-Reis D
Thu Sep 01 00:00:00 EDT 2022
Periodontal ligament fibroblasts-derived exosomes induced by PGE2 inhibit human periodontal ligament stem cells osteogenic differentiation via activating miR-34c-5p/SATB2/ERK.
Experimental cell research
Lin C,Yang Y,Wang Y,Jing H,Bai X,Hong Z,Zhang C,Gao H,Zhang L
Sat Oct 15 00:00:00 EDT 2022
Perfluorooctane sulfonate induces mitochondrial calcium overload and early hepatic insulin resistance via autophagy/detyrosinated alpha-tubulin-regulated IP3R2-VDAC1-MICU1 interaction.
The Science of the total environment
Dong Z,Wang J,Qiu T,Wu J,An Y,Shi X,Sun X,Jiang L,Liu X,Yang G,Cao J,Yao X
Wed Jun 15 00:00:00 EDT 2022
Targeting DRD2 by the antipsychotic drug, penfluridol, retards growth of renal cell carcinoma via inducing stemness inhibition and autophagy-mediated apoptosis.
Cell death & disease
Tung MC,Lin YW,Lee WJ,Wen YC,Liu YC,Chen JQ,Hsiao M,Yang YC,Chien MH
Sat Apr 23 00:00:00 EDT 2022
The m6A methyltransferase METTL3 promotes trophoblast cell invasion by regulating MYLK expression.
Placenta
Zhao J,Ding H,Ding J,Shi X,He Y,Zhu H,Yuan H,Zhang T,Zhang J
Tue Nov 01 00:00:00 EDT 2022
Effect of Houpo-Mahuang Decoction on aggravated asthma induced by cigarette smoke and the expression of TRPA1 and tight junctions in mice.
Journal of ethnopharmacology
Sun YB,Ni Y,Fan XS,Zhou LP,Yue QF,Shang EX
Fri Jul 15 00:00:00 EDT 2022
Polarity protein Par3 sensitizes breast cancer to paclitaxel by promoting cell cycle arrest.
Breast cancer research and treatment
Zhao Y,Peng H,Liang L,Li Y,Hu X,Wang B,Xu Y,Chen S
Tue Feb 01 00:00:00 EST 2022
Polarity protein Par3 sensitizes breast cancer to paclitaxel by promoting cell cycle arrest.
Breast cancer research and treatment
Zhao Y,Peng H,Liang L,Li Y,Hu X,Wang B,Xu Y,Chen S
Tue Feb 01 00:00:00 EST 2022
MRCK-Alpha and Its Effector Myosin II Regulatory Light Chain Bind ABCB4 and Regulate Its Membrane Expression.
Cells
Bruneau A,Delaunay JL,Durand-Schneider AM,Vauthier V,Ben Saad A,Aoudjehane L,El Mourabit H,Morichon R,Falguières T,Gautheron J,Housset C,Aït-Slimane T
Thu Feb 10 00:00:00 EST 2022
Porphyromonas gingivalis lipopolysaccharide induced RIPK3/MLKL-mediated necroptosis of oral epithelial cells and the further regulation in macrophage activation.
Journal of oral microbiology
Geng F,Liu J,Yin C,Zhang S,Pan Y,Sun H
Tue Mar 08 00:00:00 EST 2022
ZC3H15 promotes gastric cancer progression by targeting the FBXW7/c-Myc pathway.
Cell death discovery
Hou J,Huang P,Lan C,Geng S,Xu M,Liu Y,Chang H,Wang Z,Gu H,Wang Y,Yang G,Cui H
Fri Jan 21 00:00:00 EST 2022
A novel HDAC11 inhibitor potentiates the tumoricidal effects of cordycepin against malignant peripheral nerve sheath tumor through the Hippo signaling pathway.
American journal of cancer research
Huang PY,Shih IA,Liao YC,You HL,Lee MJ
Thu Mar 10 00:00:00 EST 2022
Two distinct males absent on the first (MOF)-containing histone acetyltransferases are involved in the epithelial-mesenchymal transition in different ways in human cells.
Cellular and molecular life sciences : CMLS
Wei T,Liu H,Zhu H,Chen W,Wu T,Bai Y,Zhang X,Miao Y,Wang F,Cai Y,Jin J
Wed Apr 13 00:00:00 EDT 2022
Predicting the Immune Microenvironment and Prognosis with a NETosis-Related lncRNA Signature in Head and Neck Squamous Cell Carcinoma.
BioMed research international
He X,Xiao Y,Liu S,Deng R,Li Z,Zhu X
Mon Sep 26 00:00:00 EDT 2022
Silencing integrin α6 enhances the pluripotency-differentiation transition in human dental pulp stem cells.
Oral diseases
Zhang W,Shen J,Zhang S,Liu X,Pan S,Li Y,Zhang L,He L,Niu Y
Fri Apr 01 00:00:00 EDT 2022
ARIH2 regulates the proliferation, DNA damage and chemosensitivity of gastric cancer cells by reducing the stability of p21 via ubiquitination.
Cell death & disease
Geng S,Peng W,Wang X,Hu X,Liang H,Hou J,Wang F,Zhao G,Lü M,Cui H
Wed Jun 22 00:00:00 EDT 2022
PGK1 represses autophagy-mediated cell death to promote the proliferation of liver cancer cells by phosphorylating PRAS40.
Cell death & disease
Zhang T,Wang Y,Yu H,Zhang T,Guo L,Xu J,Wei X,Wang N,Wu Y,Wang X,Huang L
Thu Jan 20 00:00:00 EST 2022
PGK1 represses autophagy-mediated cell death to promote the proliferation of liver cancer cells by phosphorylating PRAS40.
Cell death & disease
Zhang T,Wang Y,Yu H,Zhang T,Guo L,Xu J,Wei X,Wang N,Wu Y,Wang X,Huang L
Thu Jan 20 00:00:00 EST 2022
Implications of miR-148a-3p/p35/PTEN signaling in tau hyperphosphorylation and autoregulatory feedforward of Akt/CREB in Alzheimer's disease.
Molecular therapy. Nucleic acids
Zeng L,Jiang H,Ashraf GM,Liu J,Wang L,Zhao K,Liu M,Li Z,Liu R
Tue Mar 08 00:00:00 EST 2022
Clostridium butyricum Protects Against Pancreatic and Intestinal Injury After Severe Acute Pancreatitis via Downregulation of MMP9.
Frontiers in pharmacology
Yan Q,Jia L,Wen B,Wu Y,Zeng Y,Wang Q
Thu May 25 00:00:00 EDT 2023
Par3 promotes breast cancer invasion and migration through pull tension and protein nanoparticle-induced osmotic pressure.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Hu Y,Xie Q,Chen S,Zhao W,Zhao X,Ruan Q,Zheng Z,Zhao H,Ma T,Guo J,Li L
Tue Nov 01 00:00:00 EDT 2022
Par3 promotes breast cancer invasion and migration through pull tension and protein nanoparticle-induced osmotic pressure.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Hu Y,Xie Q,Chen S,Zhao W,Zhao X,Ruan Q,Zheng Z,Zhao H,Ma T,Guo J,Li L
Tue Nov 01 00:00:00 EDT 2022
Generation of cost-effective MXene@polydopamine-decorated chitosan nanofibrous wound dressing for promoting wound healing.
Biomaterials advances
Li H,Dai J,Yi X,Cheng F
Thu Sep 01 00:00:00 EDT 2022
IGF-1 Signaling Regulates Mitochondrial Remodeling during Myogenic Differentiation.
Nutrients
Guan X,Yan Q,Wang D,Du G,Zhou J
Wed Mar 16 00:00:00 EDT 2022
LINC01431 Promotes Histone H4R3 Methylation to Impede HBV Covalently Closed Circular DNA Transcription by Stabilizing PRMT1.
Advanced science (Weinheim, Baden-Wurttemberg, Germany)
Sun Y,Teng Y,Wang L,Zhang Z,Chen C,Wang Y,Zhang X,Xiang P,Song X,Lu J,Li N,Gao L,Liang X,Xia Y,Wu Z,Ma C
Sun May 01 00:00:00 EDT 2022
Lower expression of PDZRN3 induces endometrial carcinoma progression via the activation of canonical Wnt signaling.
Oncology letters
Li Q,Zhong J,Yang S,Liang Y
Tue Mar 01 00:00:00 EST 2022
Allopregnanolone suppresses glioblastoma survival through decreasing DPYSL3 and S100A11 expression.
The Journal of steroid biochemistry and molecular biology
Feng YH,Lim SW,Lin HY,Wang SA,Hsu SP,Kao TJ,Ko CY,Hsu TI
Sun May 01 00:00:00 EDT 2022
Gut dysbacteriosis attenuates resistance to Mycobacterium bovis infection by decreasing cyclooxygenase 2 to inhibit endoplasmic reticulum stress.
Emerging microbes & infections
Wang H,Yao J,Chen Y,Wang Y,Liu Y,Liao Y,Liang Z,Dong YH,Qu M,Ge X,Zhou X
Thu Dec 01 00:00:00 EST 2022
ESR1 Regulates the Obesity- and Metabolism-Differential Gene MMAA to Inhibit the Occurrence and Development of Hepatocellular Carcinoma.
Frontiers in oncology
Zhang Y,Cheng J,Zhong C,Xia Q,Li Y,Chen P,Fan X,Mao Q,Lin H,Hong D
Sat Jul 16 00:00:00 EDT 2022
Alarm Signal S100-Related Signature Is Correlated with Tumor Microenvironment and Predicts Prognosis in Glioma.
Disease markers
Wang LJ,Lv P,Lou Y
Mon May 23 00:00:00 EDT 2022
Anionic nanoplastic exposure induces endothelial leakiness.
Nature communications
Wei W,Li Y,Lee M,Andrikopoulos N,Lin S,Chen C,Leong DT,Ding F,Song Y,Ke PC
Sat Aug 13 00:00:00 EDT 2022
Mitochondria-Targeting Polymer Micelle of Dichloroacetate Induced Pyroptosis to Enhance Osteosarcoma Immunotherapy.
ACS nano
Jin J,Yuan P,Yu W,Lin J,Xu A,Xu X,Lou J,Yu T,Qian C,Liu B,Song J,Li L,Piao Y,Xie T,Shen Y,Tao H,Tang J
Tue Jul 26 00:00:00 EDT 2022
lncRNA HITT inhibits metastasis by attenuating Rab5-mediated endocytosis in lung adenocarcinoma.
Molecular therapy : the journal of the American Society of Gene Therapy
Wang X,Zheng S,Yang F,Zhang W,Zhao D,Xue X,Lin Q,He Y,Hu G,Hu Y
Wed Mar 02 00:00:00 EST 2022
Protective Effects of the Wenfei Buqi Tongluo Formula on the Inflammation in Idiopathic Pulmonary Fibrosis through Inhibiting the TLR4/MyD88/NF-κB Pathway.
BioMed research international
Song S,Wang J,Liu G,Ding L,Li Y,Qi H,Wei L,Zhao J,Chen T,Zhao M,Wang Z,Yang Y,Zhao D,Li X,Wang Z
Fri Apr 08 00:00:00 EDT 2022
Sinensetin attenuates IL-1β-induced cartilage damage and ameliorates osteoarthritis by regulating SERPINA3.
Food & function
Liu Z,Liu R,Wang R,Dai J,Chen H,Wang J,Li X
Mon Oct 03 00:00:00 EDT 2022
Crosstalk between acetylation and the tyrosination/detyrosination cycle of α-tubulin in Alzheimer's disease.
Frontiers in cell and developmental biology
Martínez-Hernández J,Parato J,Sharma A,Soleilhac JM,Qu X,Tein E,Sproul A,Andrieux A,Goldberg Y,Moutin MJ,Bartolini F,Peris L
Wed Sep 28 00:00:00 EDT 2022
Crosstalk between acetylation and the tyrosination/detyrosination cycle of α-tubulin in Alzheimer's disease.
Frontiers in cell and developmental biology
Martínez-Hernández J,Parato J,Sharma A,Soleilhac JM,Qu X,Tein E,Sproul A,Andrieux A,Goldberg Y,Moutin MJ,Bartolini F,Peris L
Wed Sep 28 00:00:00 EDT 2022
Crosstalk between acetylation and the tyrosination/detyrosination cycle of α-tubulin in Alzheimer's disease.
Frontiers in cell and developmental biology
Martínez-Hernández J,Parato J,Sharma A,Soleilhac JM,Qu X,Tein E,Sproul A,Andrieux A,Goldberg Y,Moutin MJ,Bartolini F,Peris L
Wed Sep 28 00:00:00 EDT 2022
Crosstalk between acetylation and the tyrosination/detyrosination cycle of α-tubulin in Alzheimer's disease.
Frontiers in cell and developmental biology
Martínez-Hernández J,Parato J,Sharma A,Soleilhac JM,Qu X,Tein E,Sproul A,Andrieux A,Goldberg Y,Moutin MJ,Bartolini F,Peris L
Wed Sep 28 00:00:00 EDT 2022
Bombesin receptor-activated protein exacerbates cisplatin-induced AKI by regulating the degradation of SIRT2.
Nephrology, dialysis, transplantation : official publication of the European Dialysis and Transplant Association - European Renal Association
Peng L,Liu D,Liu H,Xia M,Wan L,Li M,Zhao J,Tang C,Chen G,Qu X,Dong Z,Liu H
Wed Nov 23 00:00:00 EST 2022
Identification of potential key circular RNAs related to cognitive impairment after chronic constriction injury of the sciatic nerve.
Frontiers in neuroscience
Liu C,Gao R,Tang Y,Chen H,Zhang X,Sun Y,Zhao Q,Lv P,Wang H,Ye-Lehmann S,Liu J,Chen C
Wed Sep 07 00:00:00 EDT 2022
HDAC6 Inhibition Alleviates Ischemia- and Cisplatin-Induced Acute Kidney Injury by Promoting Autophagy.
Cells
Shi L,Song Z,Li C,Deng F,Xia Y,Huang J,Wu X,Zhu J
Wed Dec 07 00:00:00 EST 2022
Antidepressant-Like Effect and Mechanism of Ginsenoside Rd on Rodent Models of Depression.
Drug design, development and therapy
Li Y,Wang ML,Zhang B,Fan XX,Tang Q,Yu X,Li LN,Fan AR,Chang HS,Zhang LZ
Tue Apr 05 00:00:00 EDT 2022
Knockdown of bone morphogenetic protein 4 gene induces apoptosis and inhibits proliferation of bovine cumulus cells.
Theriogenology
Tian YQ,Li XL,Wang WJ,Hao HS,Zou HY,Pang YW,Zhao XM,Zhu HB,Du WH
Mon Aug 01 00:00:00 EDT 2022
Wnt/β-Catenin Signaling Inhibits Osteogenic Differentiation in Human Periodontal Ligament Fibroblasts.
Biomimetics (Basel, Switzerland)
Iizumi R,Honda M
Sat Dec 03 00:00:00 EST 2022
Atractylenolide III from Atractylodes macrocephala Koidz promotes the activation of brown and white adipose tissue through SIRT1/PGC-1α signaling pathway.
Phytomedicine : international journal of phytotherapy and phytopharmacology
Liu X,Huang Y,Liang X,Wu Q,Wang N,Zhou LJ,Liu WW,Ma Q,Hu B,Gao H,Cui YL,Li X,Zhao QC
Thu Sep 01 00:00:00 EDT 2022
m6A modifications regulate intestinal immunity and rotavirus infection.
eLife
Wang A,Tao W,Tong J,Gao J,Wang J,Hou G,Qian C,Zhang G,Li R,Wang D,Ren X,Zhang K,Ding S,Flavell RA,Li H,Pan W,Zhu S
Mon Jan 31 00:00:00 EST 2022
ING4 Promotes Stemness Enrichment of Human Renal Cell Carcinoma Cells Through Inhibiting DUSP4 Expression to Activate the p38 MAPK/type I IFN-Stimulated Gene Signaling Pathway.
Frontiers in pharmacology
Tang Y,Yang X,Wang Q,Huang H,Wang Q,Jiang M,Yuan C,Huang Y,Chen Y
Sat Jul 16 00:00:00 EDT 2022
FOXO acts as a positive regulator of CncC and deltamethrin tolerance in the red flour beetle, Tribolium castaneum.
Pest management science
Xu X,Zhang N,Meng X,Jiang H,Ge H,Zheng Y,Qian K,Wang J
Sun May 01 00:00:00 EDT 2022
MAD2B promotes podocyte injury through regulating Numb-dependent Notch 1 pathway in diabetic nephropathy.
International journal of biological sciences
Li MR,Lei CT,Tang H,Yin XJ,Hao Z,Qiu Y,Xie YR,Zeng JY,Su H,Zhang C
Fri Apr 15 00:00:00 EDT 2022
IL-33 in the basolateral amygdala integrates neuroinflammation into anxiogenic circuits via modulating BDNF expression.
Brain, behavior, and immunity
Zhuang X,Zhan B,Jia Y,Li C,Wu N,Zhao M,Chen N,Guo Y,Du Y,Zhang Y,Cao B,Li Y,Zhu F,Guo C,Wang Q,Li Y,Zhang L
Sun May 01 00:00:00 EDT 2022
SOX4 Mediates ATRA-Induced Differentiation in Neuroblastoma Cells.
Cancers
Zhang D,Gong B,Zhao Q,Li Z,Tan X,Hua Z
Thu Nov 17 00:00:00 EST 2022
Oncogenic role of the SOX9-DHCR24-cholesterol biosynthesis axis in IGH-BCL2+ diffuse large B-cell lymphomas.
Blood
Shen Y,Zhou J,Nie K,Cheng S,Chen Z,Wang W,Wei W,Jiang D,Peng Z,Ren Y,Zhang Y,Fan Q,Richards KL,Qi Y,Cheng J,Tam W,Ma J
Thu Jan 06 00:00:00 EST 2022
Platelet-Vesicles-Encapsulated RSL-3 Enable Anti-Angiogenesis and Induce Ferroptosis to Inhibit Pancreatic Cancer Progress.
Frontiers in endocrinology
Zhang Y,Huang Z,Cheng J,Pan H,Lin T,Shen X,Chen W,Chen Q,Gu C,Mao Q,Liang Y
Tue Apr 12 00:00:00 EDT 2022
Butyrate ameliorates skeletal muscle atrophy in diabetic nephropathy by enhancing gut barrier function and FFA2-mediated PI3K/Akt/mTOR signals.
British journal of pharmacology
Tang G,Du Y,Guan H,Jia J,Zhu N,Shi Y,Rong S,Yuan W
Sat Jan 01 00:00:00 EST 2022
HDAC9 Contributes to Serous Ovarian Cancer Progression through Regulating Epithelial-Mesenchymal Transition.
Biomedicines
Xu L,Wang J,Liu B,Fu J,Zhao Y,Yu S,Shen L,Yan X,Su J
Thu Feb 03 00:00:00 EST 2022
Matrine injection inhibits pancreatic cancer growth via modulating carbonic anhydrases- a network pharmacology-based study with in vitro validation.
Journal of ethnopharmacology
Xu PL,Cheng CS,Jiao JY,Chen H,Chen Z,Li P
Wed Apr 06 00:00:00 EDT 2022
Confinement induces oxidative damage and synaptic dysfunction in mice.
Frontiers in physiology
Li N,Wang H,Xin S,Min R,Zhang Y,Deng Y
Thu Dec 22 00:00:00 EST 2022
Melatonin reverses tumor necrosis factor-alpha-induced metabolic disturbance of human nucleus pulposus cells via MTNR1B/Gαi2/YAP signaling.
International journal of biological sciences
Qiu X,Liang T,Wu Z,Zhu Y,Gao W,Gao B,Qiu J,Wang X,Chen T,Deng Z,Li P,Chen Y,Zhou H,Peng Y,Xu C,Su P,Liang A,Huang D
Fri Apr 15 00:00:00 EDT 2022
Shenkang injection improves chronic kidney disease by inhibiting multiple renin-angiotensin system genes by blocking the Wnt/β-catenin signalling pathway.
Frontiers in pharmacology
Wang YN,Liu HJ,Ren LL,Suo P,Zou L,Zhang YM,Yu XY,Zhao YY
Wed Sep 07 00:00:00 EDT 2022
Mechanism underlying the targeted regulation of the SOD1 3'UTR by the AUF1/Dicer1/miR-155/SOD1 pathway in sodium arsenite-induced liver injury.
Ecotoxicology and environmental safety
Bi D,Shi M,Zheng D,Hu Q,Wang H,Peng L,Lou D,Zhang A,Hu Y
Thu Sep 15 00:00:00 EDT 2022
FLT4/VEGFR3 activates AMPK to coordinate glycometabolic reprogramming with autophagy and inflammasome activation for bacterial elimination.
Autophagy
Ma L,Li W,Zhang Y,Qi L,Zhao Q,Li N,Lu Y,Zhang L,Zhou F,Wu Y,He Y,Yu H,He Y,Wei B,Wang H
Wed Jun 01 00:00:00 EDT 2022
Inhibition of HDAC6 With CAY10603 Ameliorates Diabetic Kidney Disease by Suppressing NLRP3 Inflammasome.
Frontiers in pharmacology
Hou Q,Kan S,Wang Z,Shi J,Zeng C,Yang D,Jiang S,Liu Z
Tue Aug 02 00:00:00 EDT 2022
FMNL3 is Overexpressed in Tumor Tissues and Predicts an Immuno-Hot Phenotype in Pancreatic Cancer.
International journal of general medicine
Zhang Q,Nie H,Pan J,Xu H,Zhan Q
Fri Dec 02 00:00:00 EST 2022
Mitochondrial FAD shortage in SLC25A32 deficiency affects folate-mediated one-carbon metabolism.
Cellular and molecular life sciences : CMLS
Peng MZ,Shao YX,Li XZ,Zhang KD,Cai YN,Lin YT,Jiang MY,Liu ZC,Su XY,Zhang W,Jiang XL,Liu L
Tue Jun 21 00:00:00 EDT 2022
Caveolin-1 negatively regulates inflammation and fibrosis in silicosis.
Journal of cellular and molecular medicine
He R,Yuan X,Lv X,Liu Q,Tao L,Meng J
Sat Jan 01 00:00:00 EST 2022
OTUB1 suppresses Hippo signaling via modulating YAP protein in gastric cancer.
Oncogene
Yan C,Yang H,Su P,Li X,Li Z,Wang D,Zang Y,Wang T,Liu Z,Bao Z,Dong S,Zhuang T,Zhu J,Ding Y
Tue Nov 01 00:00:00 EDT 2022
N6 -Methyladenosine-Modified CBX1 Regulates Nasopharyngeal Carcinoma Progression Through Heterochromatin Formation and STAT1 Activation.
Advanced science (Weinheim, Baden-Wurttemberg, Germany)
Zhao Y,Huang S,Tan X,Long L,He Q,Liang X,Bai J,Li Q,Lin J,Li Y,Liu N,Ma J,Chen Y
Thu Dec 01 00:00:00 EST 2022
Downregulation of HINFP induces senescence-associated secretory phenotype to promote metastasis in a non-cell-autonomous manner in bladder cancer.
Oncogene
Zheng X,Liu Z,Zhong J,Zhou L,Chen J,Zheng L,Li Z,Zhang R,Pan J,Wu Y,Liu Z,Kang T
Fri Jul 01 00:00:00 EDT 2022
ZC3H15 promotes glioblastoma progression through regulating EGFR stability.
Cell death & disease
Hou J,Xu M,Gu H,Pei D,Liu Y,Huang P,Chang H,Cui H
Thu Jan 13 00:00:00 EST 2022
De novo non-synonymous DPYSL2 (CRMP2) variants in two patients with intellectual disabilities and documentation of functional relevance through zebrafish rescue and cellular transfection experiments.
Human molecular genetics
Suzuki H,Li S,Tokutomi T,Takeuchi C,Takahashi M,Yamada M,Okuno H,Miya F,Takenouchi T,Numabe H,Kosaki K,Ohshima T
Fri Dec 16 00:00:00 EST 2022
STRIPAK regulation of katanin microtubule severing in the Caenorhabditis elegans embryo.
Genetics
Lu T,Smit RB,Soueid H,Mains PE
Thu May 05 00:00:00 EDT 2022
Identification of a De Novo Heterozygous Missense ACTB Variant in Baraitser-Winter Cerebrofrontofacial Syndrome.
Frontiers in genetics
Nie K,Huang J,Liu L,Lv H,Chen D,Fan W
Wed Apr 13 00:00:00 EDT 2022
KCTD10 regulates brown adipose tissue thermogenesis and metabolic function via Notch signaling.
The Journal of endocrinology
Ye MS,Luo L,Guo Q,Su T,Cheng P,Huang Y
Thu Jan 13 00:00:00 EST 2022
DNA 6mA Demethylase ALKBH1 Orchestrates Fatty Acid Metabolism and Suppresses Diet-Induced Hepatic Steatosis.
Cellular and molecular gastroenterology and hepatology
Luo L,Liu Y,Nizigiyimana P,Ye M,Xiao Y,Guo Q,Su T,Luo X,Huang Y,Zhou H
Tue Nov 22 00:00:00 EST 2022
High expression of small nucleolar RNA host gene 3 predicts poor prognosis and promotes bone metastasis in prostate cancer by activating transforming growth factor-beta signaling.
Bioengineered
Xi X,Hu Z,Wu Q,Hu K,Cao Z,Zhou J,Liao J,Zhang Z,Hu Y,Zhong X,Bao Y
Sat Jan 01 00:00:00 EST 2022
Prinsepiae Nux Extract Activates NRF2 Activity and Protects UVB-Induced Damage in Keratinocyte.
Antioxidants (Basel, Switzerland)
Wang SH,Chen YS,Lai KH,Lu CK,Chang HS,Wu HC,Yen FL,Chen LY,Lee JC,Yen CH
Mon Sep 05 00:00:00 EDT 2022
UPF1 contributes to the maintenance of endometrial cancer stem cell phenotype by stabilizing LINC00963.
Cell death & disease
Chen H,Ma J,Kong F,Song N,Wang C,Ma X
Tue Mar 22 00:00:00 EDT 2022
Artemisinin relieves myocardial ischemia-reperfusion injury via modulating miR-29b-3p and hemicentin 1.
Frontiers in pharmacology
Han J,Zhang Z,Zhang Z,Yang S
Mon Mar 20 00:00:00 EDT 2023
Box C/D snoRNA SNORD89 influences the occurrence and development of endometrial cancer through 2'-O-methylation modification of Bim.
Cell death discovery
Bao HJ,Chen X,Liu X,Wu W,Li QH,Xian JY,Zhao Y,Chen S
Tue Jul 05 00:00:00 EDT 2022
Eupalinolide B attenuates lipopolysaccharide-induced acute lung injury through inhibition of NF-κB and MAPKs signaling by targeting TAK1 protein.
International immunopharmacology
Yang L,Chen H,Hu Q,Liu L,Yuan Y,Zhang C,Tang J,Shen X
Sat Oct 01 00:00:00 EDT 2022
PCGF6 controls neuroectoderm specification of human pluripotent stem cells by activating SOX2 expression.
Nature communications
Lan X,Ding S,Zhang T,Yi Y,Li C,Jin W,Chen J,Liang K,Wang H,Jiang W
Sat Aug 06 00:00:00 EDT 2022
TET2-BCLAF1 transcription repression complex epigenetically regulates the expression of colorectal cancer gene Ascl2 via methylation of its promoter.
The Journal of biological chemistry
Shang Y,Jiang T,Ran L,Hu W,Wu Y,Ye J,Peng Z,Chen L,Wang R
Fri Jul 01 00:00:00 EDT 2022
Myostain is involved in ginsenoside Rb1-mediated anti-obesity.
Pharmaceutical biology
Li HS,Kuang JY,Liu GJ,Wu WJ,Yin XL,Li HD,Wang L,Qin T,Zhang WC,Sun YY
Thu Dec 01 00:00:00 EST 2022
Electroacupuncture-Regulated miR-34a-3p/PDCD6 Axis Promotes Post-Spinal Cord Injury Recovery in Both In Vitro and In Vivo Settings.
Journal of immunology research
Ma L,Ma L,Yang Y,Chen T,Wang L,Deng Q
Fri Sep 23 00:00:00 EDT 2022
Electroacupuncture-Regulated miR-34a-3p/PDCD6 Axis Promotes Post-Spinal Cord Injury Recovery in Both In Vitro and In Vivo Settings.
Journal of immunology research
Ma L,Ma L,Yang Y,Chen T,Wang L,Deng Q
Fri Sep 23 00:00:00 EDT 2022
DJ-1 promotes osteosarcoma progression through activating CDK4/RB/E2F1 signaling pathway.
Frontiers in oncology
Han Z,Wang L,Wang D,Zhang L,Bi Y,Zheng X,Liu W,Bai G,Wang Z,Wan W,Ma Y,Cai X,Liu T,Jia Q
Mon Nov 06 00:00:00 EST 2023
Drebrin promotes lung adenocarcinoma cell migration through inducing integrin β1 endocytosis.
Biochemical and biophysical research communications
Yu Q,Tang F,Cao F,Tan X,Weng L,Sun L
Sat Nov 19 00:00:00 EST 2022
Nedd4-2 Haploinsufficiency in Mice Impairs the Ubiquitination of Rer1 and Increases the Susceptibility to Endoplasmic Reticulum Stress and Seizures.
Frontiers in molecular neuroscience
Liu X,Zhang L,Zhang H,Liang X,Zhang B,Tu J,Zhao Y
Sat Jul 16 00:00:00 EDT 2022
Chemerin Regulates the Proliferation and Migration of Pulmonary Arterial Smooth Muscle Cells via the ERK1/2 Signaling Pathway.
Frontiers in pharmacology
Peng L,Chen Y,Li Y,Feng P,Zheng Y,Dong Y,Yang Y,Wang R,Li A,Yan J,Shang F,Tang P,Chen D,Gao Y,Huang W
Tue Apr 05 00:00:00 EDT 2022
RBCK1 is an endogenous inhibitor for triple negative breast cancer via hippo/YAP axis.
Cell communication and signaling : CCS
Li Z,Su P,Ding Y,Gao H,Yang H,Li X,Yang X,Xia Y,Zhang C,Fu M,Wang D,Zhang Y,Zhuo S,Zhu J,Zhuang T
Mon Oct 24 00:00:00 EDT 2022
Inhibition of nuclear deacetylase Sirtuin-1 induces mitochondrial acetylation and calcium overload leading to cell death.
Redox biology
Sun Y,Yang YM,Hu YY,Ouyang L,Sun ZH,Yin XF,Li N,He QY,Wang Y
Fri Jul 01 00:00:00 EDT 2022
LncRNA LINC00281/Annexin A2 Regulates Vascular Smooth Muscle Cell Phenotype Switching via the Nuclear Factor-Kappa B Signaling Pathway.
Journal of cardiovascular translational research
Cheng L,Wang H,Maboh R,Mao G,Wu X,Chen H
Sat Oct 01 00:00:00 EDT 2022
N6-methyladenosine (m6A) in 18S rRNA promotes fatty acid metabolism and oncogenic transformation.
Nature metabolism
Peng H,Chen B,Wei W,Guo S,Han H,Yang C,Ma J,Wang L,Peng S,Kuang M,Lin S
Mon Aug 01 00:00:00 EDT 2022
CEND1 deficiency induces mitochondrial dysfunction and cognitive impairment in Alzheimer's disease.
Cell death and differentiation
Xie W,Guo D,Li J,Yue L,Kang Q,Chen G,Zhou T,Wang H,Zhuang K,Leng L,Li H,Chen Z,Gao W,Zhang J
Thu Dec 01 00:00:00 EST 2022
Laparotomy-Induced Peripheral Inflammation Activates NR2B Receptors on the Brain Mast Cells and Results in Neuroinflammation in a Vagus Nerve-Dependent Manner.
Frontiers in cellular neuroscience
Yang J,Dong HQ,Liu YH,Ji MH,Zhang X,Dai HY,Sun ZC,Liu L,Zhou J,Sha HH,Qian YN,Li QG,Yao H,Li NN
Tue Mar 01 00:00:00 EST 2022
Blockade of TMEM16A protects against renal fibrosis by reducing intracellular Cl- concentration.
British journal of pharmacology
Li XL,Liu J,Chen XS,Cheng LM,Liu WL,Chen XF,Li YJ,Guan YY,Zeng X,Du YH
Wed Jun 01 00:00:00 EDT 2022
SIRT6 overexpression retards renal interstitial fibrosis through targeting HIPK2 in chronic kidney disease.
Frontiers in pharmacology
Li X,Li W,Zhang Z,Wang W,Huang H
Fri Sep 30 00:00:00 EDT 2022
Microtubule associated protein 4 phosphorylation-induced epithelial-to-mesenchymal transition of podocyte leads to proteinuria in diabetic nephropathy.
Cell communication and signaling : CCS
Li L,Feng Y,Zhang J,Zhang Q,Ren J,Sun C,Li S,Lei X,Luo G,Hu J,Huang Y
Thu Jul 28 00:00:00 EDT 2022
Monitoring Protein Import into the Endoplasmic Reticulum in Living Cells with Proximity Labeling.
ACS chemical biology
Lyu Z,Sycks MM,Espinoza MF,Nguyen KK,Montoya MR,Galapate CM,Mei L,Genereux JC
Fri Jul 15 00:00:00 EDT 2022
The Ancient and Evolved Mouse Sperm-Associated Antigen 6 Genes Have Different Biologic Functions In Vivo.
Cells
Yap YT,Li W,Zhou Q,Haj-Diab S,Chowdhury DD,Vaishnav A,Harding P,Williams DC,Edwards BF,Strauss JF,Zhang Z
Thu Jan 20 00:00:00 EST 2022
A High Hepatic Uptake of Conjugated Bile Acids Promotes Colorectal Cancer-Associated Liver Metastasis.
Cells
Zheng Z,Wei J,Hou X,Jia F,Zhang Z,Guo H,Yuan F,He F,Ke Z,Wang Y,Zhao L
Mon Nov 28 00:00:00 EST 2022
Nardilysin in adipocytes regulates UCP1 expression and body temperature homeostasis.
Scientific reports
Saijo S,Ohno M,Iwasaki H,Matsuda S,Nishi K,Hiraoka Y,Ide N,Kimura T,Nishi E
Wed Mar 02 00:00:00 EST 2022
Circ-CREBBP inhibits sperm apoptosis via the PI3K-Akt signaling pathway by sponging miR-10384 and miR-143-3p.
Communications biology
Ding N,Zhang Y,Huang M,Liu J,Wang C,Zhang C,Cao J,Zhang Q,Jiang L
Wed Dec 07 00:00:00 EST 2022
p-hydroxy benzaldehyde revitalizes the microenvironment of peri-infarct cortex in rats after cerebral ischemia-reperfusion.
Phytomedicine : international journal of phytotherapy and phytopharmacology
Yuan Y,Liu L,Du Y,Fan R,Zhang R,Zhou N
Sat Oct 01 00:00:00 EDT 2022
HOXA5 inhibits the proliferation of extrahepatic cholangiocarcinoma cells by enhancing MXD1 expression and activating the p53 pathway.
Cell death & disease
Xiong F,Liu W,Wang X,Wu G,Wang Q,Guo T,Huang W,Wang B,Chen Y
Tue Sep 27 00:00:00 EDT 2022
ATRA-mediated-crosstalk between stellate cells and Kupffer cells inhibits autophagy and promotes NLRP3 activation in acute liver injury.
Cellular signalling
Yu Z,Xie X,Su X,Lv H,Song S,Liu C,You Y,Tian M,Zhu L,Wang L,Qi J,Zhu Q
Sun May 01 00:00:00 EDT 2022
Time-dependent expression of ryanodine receptors in sea urchin eggs, zygotes and early embryos.
Zygote (Cambridge, England)
Percivale G,Angelini C,Falugi C,Picco C,Prestipino G
Fri Apr 01 00:00:00 EDT 2022
Brain aging is faithfully modelled in organotypic brain slices and accelerated by prions.
Communications biology
Liu Y,Senatore A,Sorce S,Nuvolone M,Guo J,Gümüş ZH,Aguzzi A
Wed Jun 08 00:00:00 EDT 2022
PGC-1β maintains mitochondrial metabolism and restrains inflammatory gene expression.
Scientific reports
Guak H,Sheldon RD,Beddows I,Vander Ark A,Weiland MJ,Shen H,Jones RG,St-Pierre J,Ma EH,Krawczyk CM
Mon Sep 26 00:00:00 EDT 2022
SIRT6 protects vascular smooth muscle cells from osteogenic transdifferentiation via Runx2 in chronic kidney disease.
The Journal of clinical investigation
Li W,Feng W,Su X,Luo D,Li Z,Zhou Y,Zhu Y,Zhang M,Chen J,Liu B,Huang H
Tue Jan 04 00:00:00 EST 2022
Cucurbitacin E inhibits cellular proliferation and induces apoptosis in melanoma by suppressing HSDL2 expression.
Chinese medicine
Liu WB,Wang HL,Chen L,Tang B,Ke G,Wang S,Sun YQ,Ma J,Lyu DL
Tue Feb 22 00:00:00 EST 2022
Impaired Bestrophin Channel Activity in an iPSC-RPE Model of Best Vitelliform Macular Dystrophy (BVMD) from an Early Onset Patient Carrying the P77S Dominant Mutation.
International journal of molecular sciences
Navinés-Ferrer A,Ruiz-Nogales S,Navarro R,Pomares E
Mon Jul 04 00:00:00 EDT 2022
Cullin 4b-RING ubiquitin ligase targets IRGM1 to regulate Wnt signaling and intestinal homeostasis.
Cell death and differentiation
Fan Y,Huo X,Guo B,Zhang X,Yang Y,Lian J,Meng X,Shao Y,Zou Y,Guo H,Wang H,Sun G,Dou H,Wang J,Shao C,Gong Y,Hu H
Thu Sep 01 00:00:00 EDT 2022
Quinoline alkaloids with anti-inflammatory activity from Zanthoxylum avicennae.
Organic & biomolecular chemistry
Ji KL,Liu W,Yin WH,Li JY,Yue JM
Thu May 26 00:00:00 EDT 2022
C/D box small nucleolar RNA SNORD104 promotes endometrial cancer by regulating the 2'-O-methylation of PARP1.
Journal of translational medicine
Lu B,Chen X,Liu X,Chen J,Qin H,Chen S,Zhao Y
Sat Dec 24 00:00:00 EST 2022
Fatty acid-binding protein 5 aggravates pulmonary artery fibrosis in pulmonary hypertension secondary to left heart disease via activating wnt/β-catenin pathway.
Journal of advanced research
Lei Q,Yu Z,Li H,Cheng J,Wang Y
Thu Sep 01 00:00:00 EDT 2022
The transcription factor KLF14 attenuates LPS-induced acute lung injury by ameliorating apoptosis of alveolar epithelial cells in mice.
Molecular immunology
Liu L,Yuan Y,Zhou Y,Yao L,Li J,Chen F
Thu Dec 01 00:00:00 EST 2022
Nrf2 and NF-κB/NLRP3 inflammasome pathways are involved in Prototheca bovis infections of mouse mammary gland tissue and mammary epithelial cells.
Free radical biology & medicine
Zhao W,Deng Z,Barkema HW,Xu M,Gao J,Liu G,Lin Y,Kastelic JP,Han B
Sun May 01 00:00:00 EDT 2022
Ciliary transition zone proteins coordinate ciliary protein composition and ectosome shedding.
Nature communications
Wang L,Wen X,Wang Z,Lin Z,Li C,Zhou H,Yu H,Li Y,Cheng Y,Chen Y,Lou G,Pan J,Cao M
Sat Jul 09 00:00:00 EDT 2022
The Expression and Role Analysis of Methylation-Regulated Differentially Expressed Gene UBE2C in Pan-Cancer, Especially for HGSOC.
Cancers
Li J,Sun Y,Zhi X,Li Q,Yao L,Chen M
Sat Jun 25 00:00:00 EDT 2022
A pyroptosis-related gene model and its correlation with the microenvironment of lung adenocarcinoma: A bioinformatics analysis and experimental verification.
Frontiers in genetics
Dong Y,Yi L,Song Q,Yao Y
Tue Nov 29 00:00:00 EST 2022
Moderate heart rate reduction promotes cardiac regeneration through stimulation of the metabolic pattern switch.
Cell reports
Tan J,Yang M,Wang H,Shen C,Wu M,Xu H,Wu Y,Li Y,Li X,Huang T,Deng S,Yang Z,Gao S,Li H,Zhou J,Chen H,Cao N,Cai W
Tue Mar 08 00:00:00 EST 2022
Human cytomegalovirus UL24 and UL43 products participate in SAMHD1 subcellular localization.
Virusdisease
Sleman S,Najmuldeen H,Hao H,Jalal P,Saeed N,Othman D,Qian Z
Thu Dec 01 00:00:00 EST 2022
mTORC1 Mediates the Processes of Lysine Regulating Satellite Cells Proliferation, Apoptosis, and Autophagy.
Metabolites
Liu M,Yue Z,Zhang B,Li F,Liu L,Li F
Thu Aug 25 00:00:00 EDT 2022
Impaired KDM2B-mediated PRC1 recruitment to chromatin causes defective neural stem cell self-renewal and ASD/ID-like behaviors.
iScience
Gao Y,Duque-Wilckens N,Aljazi MB,Moeser AJ,Mias GI,Robison AJ,Zhang Y,He J
Fri Feb 18 00:00:00 EST 2022
HDAC6 Inhibition Alleviates Anesthesia and Surgery-Induced Less Medial Prefrontal-Dorsal Hippocampus Connectivity and Cognitive Impairment in Aged Rats.
Molecular neurobiology
Chen J,Liu S,Wang X,Huang J,Phillips J,Ma D,Ouyang W,Tong J
Sat Oct 01 00:00:00 EDT 2022
Aldehyde dehydrogenase 2 and PARP1 interaction modulates hepatic HDL biogenesis by LXRα-mediated ABCA1 expression.
JCI insight
Li L,Zhong S,Li R,Liang N,Zhang L,Xia S,Xu X,Chen X,Chen S,Tao Y,Yin H
Fri Apr 08 00:00:00 EDT 2022
Targeting polarized phenotype of microglia via IL6/JAK2/STAT3 signaling to reduce NSCLC brain metastasis.
Signal transduction and targeted therapy
Jin Y,Kang Y,Wang M,Wu B,Su B,Yin H,Tang Y,Li Q,Wei W,Mei Q,Hu G,Lukacs-Kornek V,Li J,Wu K,Yuan X,Wang W
Wed Feb 23 00:00:00 EST 2022
The Photoperiod Regulates Granulosa Cell Apoptosis through the FSH-Nodal/ALK7 Signaling Pathway in Phodopus sungorus.
Animals : an open access journal from MDPI
Qi Y,Xue HL,Zheng YJ,Yin YF,Xu WL,Xu JH,Wu M,Chen L,Xu LX
Fri Dec 16 00:00:00 EST 2022
Loss of NPPA-AS1 promotes heart regeneration by stabilizing SFPQ-NONO heteromer-induced DNA repair.
Basic research in cardiology
Fu W,Ren H,Shou J,Liao Q,Li L,Shi Y,Jose PA,Zeng C,Wang WE
Sat Mar 05 00:00:00 EST 2022
Overexpression of sFlt-1 represses ox-LDL-induced injury of HUVECs by activating autophagy via PI3K/AKT/mTOR pathway.
Microvascular research
Zhou YH,Tang YZ,Guo LY,Zheng LL,Zhang D,Yang CY,Wang W
Sat Jan 01 00:00:00 EST 2022
Low STING expression promotes endometrial stromal cell invasion and migration via the STING/IRF-3/IFN-β1 pathway in eutopic endometrium of women with endometriosis.
Gynecological endocrinology : the official journal of the International Society of Gynecological Endocrinology
Xu Z,Zhao H,Yue C,Zhang L,Li M,Du Y,Zhang N,Yan L
Thu Dec 01 00:00:00 EST 2022
Bifidobacterium spp. and their metabolite lactate protect against acute pancreatitis via inhibition of pancreatic and systemic inflammatory responses.
Gut microbes
Li H,Xie J,Guo X,Yang G,Cai B,Liu J,Yue M,Tang Y,Wang G,Chen S,Guo J,Qi X,Wang D,Zheng H,Liu W,Yu H,Wang C,Zhu SJ,Guo F
Thu Oct 06 00:00:00 EDT 2022
In vitro and in vivo assay of the ER lipid scramblase TMEM41B.
STAR protocols
Wu L,Liu L,Xu B,Huang D,Chen XW
Fri Jun 17 00:00:00 EDT 2022
White tea alleviates non-alcoholic fatty liver disease by regulating energy expenditure and lipid metabolism.
Gene
Li N,Zhou X,Wang J,Chen J,Lu Y,Sun Y,Song Y,Tan X,Xie G,Chen Y,Zhang L
Sat Jul 30 00:00:00 EDT 2022
PKC-ζ Aggravates Doxorubicin-Induced Cardiotoxicity by Inhibiting Wnt/β-Catenin Signaling.
Frontiers in pharmacology
Cao YJ,Li JY,Wang PX,Lin ZR,Yu WJ,Zhang JG,Lu J,Liu PQ
Fri Mar 04 00:00:00 EST 2022
Design and synthesis of Aza-boeravinone derivatives as potential novel topoisomerase I inhibitors.
Bioorganic chemistry
Zhou Y,Bai YP,Zhang M,Gao JM,Yang CJ,Zhang ZJ,Deng N,Li L,Liu YQ,Xu CR
Sun May 01 00:00:00 EDT 2022
The Role of Sirtuin 1 in Palmitic Acid-Induced Endoplasmic Reticulum Stress in Cardiac Myoblasts.
Life (Basel, Switzerland)
Yang HY,Chen JY,Huo YN,Yu PL,Lin PZ,Hsu SC,Huang SM,Tsai CS,Lin CY
Wed Jan 26 00:00:00 EST 2022
The phosphatase of regenerating liver-3 protein (PRL-3) promotes glioma cell invasiveness by interacting with β3 -tubulin.
Bioengineered
Ge Z,Gu T,Zhang L,Fan Q,Ma L,Fang N
Tue Feb 01 00:00:00 EST 2022
miR21 modulates the Hippo signaling pathway via interference with PP2A Bβ to inhibit trophoblast invasion and cause preeclampsia.
Molecular therapy. Nucleic acids
Hu M,Zheng Y,Liao J,Wen L,Cheng J,Huang J,Huang B,Lin L,Long Y,Wu Y,Ye X,Fu Y,Qi H,Baker PN,Tong C
Tue Dec 13 00:00:00 EST 2022
Design, Synthesis, and Biological Evaluation of Androgen Receptor Degrading and Antagonizing Bifunctional Steroidal Analogs for the Treatment of Advanced Prostate Cancer.
Journal of medicinal chemistry
Wang A,Luo X,Wang Y,Meng X,Lu Z,Yang Y
Thu Sep 22 00:00:00 EDT 2022
The Ion Channel Gene KCNAB2 Is Associated with Poor Prognosis and Loss of Immune Infiltration in Lung Adenocarcinoma.
Cells
Lyu Y,Wang Q,Liang J,Zhang L,Zhang H
Mon Oct 31 00:00:00 EDT 2022
Grass Carp Reovirus triggers autophagy enhancing virus replication via the Akt/mTOR pathway.
Fish & shellfish immunology
Zhu M,Zhang Y,Pan J,Tong X,Zhang X,Hu X,Gong C
Thu Sep 01 00:00:00 EDT 2022
Suppressing MDSC Infiltration in Tumor Microenvironment Serves as an Option for Treating Ovarian Cancer Metastasis.
International journal of biological sciences
Li Y,Zhang Q,Wu M,Zhang P,Huang L,Ai X,Yang Z,Shen Q,Wang Y,Wang P,Zhou S,He ML
Tue Jul 12 00:00:00 EDT 2022
Oxidative stress modulates expression of immune checkpoint genes via activation of AhR signaling.
Toxicology and applied pharmacology
Kou Z,Yang R,Lee E,Cuddapah S,Choi BH,Dai W
Thu Dec 15 00:00:00 EST 2022
MACF1 promotes osteoblastic cell migration by regulating MAP1B through the GSK3beta/TCF7 pathway.
Bone
Su P,Tian Y,Yin C,Wang X,Li D,Yang C,Pei J,Deng X,King S,Li Y,Qian A
Sat Jan 01 00:00:00 EST 2022
GPR12 Inhibits Apoptosis in Epithelial Ovarian Cancer via the Activation of ERK1/2 Signaling.
Frontiers in oncology
Wang L,Yang D,Zhang Y,Jiao Y
Sun Jul 31 00:00:00 EDT 2022
TOPK Activation Exerts Protective Effects on Cisplatin-induced Acute Kidney Injury.
Current medical science
Zhang H,Dong QQ,Shu HP,Tu YC,Liao QQ,Yao LJ
Mon Aug 01 00:00:00 EDT 2022
Qiangli Wuhu mixture alleviates LPS-induced pneumonia by inhibiting the TLR4/NF-κB/NLRP3 pathway: a study based on network pharmacology.
Pharmaceutical biology
Tian J,Wang XL,Wang LC,Chen F,Tian Y,Ma L,Pan CY,Wang YP
Thu Dec 01 00:00:00 EST 2022
The hypoxia-inducible factor prolyl hydroxylase inhibitor FG4592 promotes natriuresis through upregulation of COX2 in the renal medulla.
Hypertension research : official journal of the Japanese Society of Hypertension
Guan N,Zhang M,Gong WY,Mao XY,Yang SS,Hao CM
Sun May 01 00:00:00 EDT 2022
Transcriptional Repression of Aerobic Glycolysis by OVOL2 in Breast Cancer.
Advanced science (Weinheim, Baden-Wurttemberg, Germany)
Zhang X,Luo F,Luo S,Li L,Ren X,Lin J,Liang Y,Ma C,Ding L,Zhang D,Ye T,Lin Y,Jin B,Gao S,Ye Q
Thu Sep 01 00:00:00 EDT 2022
RNF6 promotes the migration and invasion of breast cancer by promoting the ubiquitination and degradation of MST1.
Experimental and therapeutic medicine
Huang Y,Zou Y,Jie Z
Tue Feb 01 00:00:00 EST 2022
Metabolic Reprogramming in Response to Alterations of Mitochondrial DNA and Mitochondrial Dysfunction in Gastric Adenocarcinoma.
International journal of molecular sciences
Chang TC,Lee HT,Pan SC,Cho SH,Cheng C,Ou LH,Lin CI,Lin CS,Wei YH
Sun Feb 06 00:00:00 EST 2022
Cholesterol determines the cytosolic entry and seeded aggregation of tau.
Cell reports
Tuck BJ,Miller LVC,Katsinelos T,Smith AE,Wilson EL,Keeling S,Cheng S,Vaysburd MJ,Knox C,Tredgett L,Metzakopian E,James LC,McEwan WA
Tue May 03 00:00:00 EDT 2022
Gentiopicroside Ameliorates Diabetic Renal Tubulointerstitial Fibrosis via Inhibiting the AT1R/CK2/NF-κB Pathway.
Frontiers in pharmacology
Xu Z,Zhang M,Wang Y,Chen R,Xu S,Sun X,Yang Y,Lin Z,Wang S,Huang H
Sat Jul 16 00:00:00 EDT 2022
Critical role of guanylate binding protein 5 in tumor immune microenvironment and predictive value of immunotherapy response.
Frontiers in genetics
Li X,Song D,Su S,He X,Cao F,Yang C,Li K,Huang S,Li C,Wang C,Zhang A,Pang P,Zheng Y
Wed Oct 19 00:00:00 EDT 2022
Alkbh1-mediated DNA N6-methyladenine modification regulates bone marrow mesenchymal stem cell fate during skeletal aging.
Cell proliferation
Cai GP,Liu YL,Luo LP,Xiao Y,Jiang TJ,Yuan J,Wang M
Tue Feb 01 00:00:00 EST 2022
STAT6 Upregulates NRP1 Expression in Endothelial Cells and Promotes Angiogenesis.
Frontiers in oncology
Gao P,Ren G,Liang J,Liu J
Sat Jul 16 00:00:00 EDT 2022
Narciclasine inhibits phospholipase A2 and regulates phospholipid metabolism to ameliorate psoriasis-like dermatitis.
Frontiers in immunology
Kong Y,Jiang J,Huang Y,Liu X,Jin Z,Li L,Wei F,Liu X,Yin J,Zhang Y,Tong Q,Chen H
Mon Jan 30 00:00:00 EST 2023
Aurora kinase blockade drives de novo addiction of cervical squamous cell carcinoma to druggable EGFR signalling.
Oncogene
Komatsu M,Nakamura K,Takeda T,Chiwaki F,Banno K,Aoki D,Takeshita F,Sasaki H
Fri Apr 01 00:00:00 EDT 2022
Hypoxic training upregulates mitochondrial turnover and angiogenesis of skeletal muscle in mice.
Life sciences
Zhao YC,Guo W,Gao BH
Tue Feb 15 00:00:00 EST 2022
Tumor-targeted dual-starvation therapy based on redox-responsive micelle nanosystem with co-loaded LND and BPTES.
Materials today. Bio
Fu Z,Du H,Meng S,Yao M,Zhao P,Li X,Zheng X,Yuan Z,Yang H,Cai K,Dai L
Thu Dec 01 00:00:00 EST 2022
Improvement of anticancer effect of berberine by salt formation modifications.
Phytomedicine : international journal of phytotherapy and phytopharmacology
Xu F,Liu M,Liao Y,Zhou Y,Zhang P,Zeng Y,Liu Z
Thu Sep 01 00:00:00 EDT 2022
TERC suppresses PD-L1 expression by downregulating RNA binding protein HuR.
Science China. Life sciences
Jin H,Chen Y,Ren J,Huang J,Zhao Y,Liu H
Thu Dec 01 00:00:00 EST 2022
Induction of zinc finger protein RNF6 auto-ubiquitination for the treatment of myeloma and chronic myeloid leukemia.
The Journal of biological chemistry
Zhuang H,Ren Y,Mao C,Zhong Y,Zhang Z,Cao B,Zhang Y,Huang J,Xu G,Huang Z,Xu Y,Mao X
Thu Sep 01 00:00:00 EDT 2022
SUMO2-mediated SUMOylation of SH3GLB1 promotes ionizing radiation-induced hypertrophic cardiomyopathy through mitophagy activation.
European journal of pharmacology
Gao A,Zou J,Mao Z,Zhou H,Zeng G
Sun Jun 05 00:00:00 EDT 2022
Proteomic characterization of gastric cancer response to chemotherapy and targeted therapy reveals new therapeutic strategies.
Nature communications
Li Y,Xu C,Wang B,Xu F,Ma F,Qu Y,Jiang D,Li K,Feng J,Tian S,Wu X,Wang Y,Liu Y,Qin Z,Liu Y,Qin J,Song Q,Zhang X,Sujie A,Huang J,Liu T,Shen K,Zhao JY,Hou Y,Ding C
Thu Sep 29 00:00:00 EDT 2022
The Transcription of ZIP9 Is Associated With the Macrophage Polarization and the Pathogenesis of Hepatocellular Carcinoma.
Frontiers in immunology
Gou Y,Yang D,Tian T,Zhu X,Zhang R,Ren J,Tu D,Luo Y,Miao Y,Zhao H,Wang Y,Wei B
Fri Apr 08 00:00:00 EDT 2022
eEF2K promotes PD-L1 stabilization through inactivating GSK3β in melanoma.
Journal for immunotherapy of cancer
Chen X,Wang K,Jiang S,Sun H,Che X,Zhang M,He J,Wen Y,Liao M,Li X,Zhou X,Song J,Ren X,Yi W,Yang J,Chen X,Yin M,Cheng Y
Tue Mar 01 00:00:00 EST 2022
Corilagin Restrains NLRP3 Inflammasome Activation and Pyroptosis through the ROS/TXNIP/NLRP3 Pathway to Prevent Inflammation.
Oxidative medicine and cellular longevity
Luo T,Zhou X,Qin M,Lin Y,Lin J,Chen G,Liu A,Ouyang D,Chen D,Pan H
Fri Oct 28 00:00:00 EDT 2022
Peptide PDHPS1 Inhibits Ovarian Cancer Growth through Disrupting YAP Signaling.
Molecular cancer therapeutics
Pan X,Geng Z,Li J,Li X,Zhang M,Wang X,Cong Y,Huang K,Xu J,Jia X
Tue Jul 05 00:00:00 EDT 2022
PGM3 regulates beta-catenin activity to promote colorectal cancer cell progression.
Experimental biology and medicine (Maywood, N.J.)
Zhang N,Liu S,Xu J,Ning T,Xie S,Min L,Zhu S,Zhang S,Zhu S
Thu Sep 01 00:00:00 EDT 2022
Hypoxic Training Ameliorates Skeletal Muscle Microcirculation Vascular Function in a Sirt3-Dependent Manner.
Frontiers in physiology
Ma C,Zhao Y,Ding X,Gao B
Fri Aug 05 00:00:00 EDT 2022
Slit guidance ligand 2 promotes the inflammatory response of periodontitis through activation of the NF-κB signaling pathway.
Journal of periodontal research
Sun H,Li Z,Fan C,Liu S,Yan K,Huang G,Li S
Wed Jun 01 00:00:00 EDT 2022
A novel isoform of hydroxyacyl-CoA dehydrogenase inhibits cell proliferation.
Biochemical and biophysical research communications
Xia G,Gao Y,Wu C,Pan H,Hou J,Su J,Wei S,Gao X
Sat May 28 00:00:00 EDT 2022
Angiotensin II induces podocyte metabolic reprogramming from glycolysis to glycerol-3-phosphate biosynthesis.
Cellular signalling
Luo Z,Chen Z,Zhu Z,Hao Y,Feng J,Luo Q,Zhang Z,Yang X,Hu J,Liang W,Ding G
Tue Nov 01 00:00:00 EDT 2022
Inhibitory effect of aptamer-carbon dot nanomaterial-siRNA complex on the metastasis of hepatocellular carcinoma cells by interfering with FMRP.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V
Zhao X,Yang J,Zhang J,Wang X,Chen L,Zhang C,Shen Z
Sun May 01 00:00:00 EDT 2022
Silencing GOLGA8B inhibits cell invasion and metastasis by suppressing STAT3 signaling pathway in lung squamous cell carcinoma.
Clinical science (London, England : 1979)
Li Z,Li Y,Li N,Shen L,Liu A
Fri Jun 17 00:00:00 EDT 2022
Identification of MYEOV-Associated Gene Network as a Potential Therapeutic Target in Pancreatic Cancer.
Cancers
Chen Y,Wang J,Guo Q,Li X,Zou X
Fri Nov 04 00:00:00 EDT 2022
CIRBP-OGFR axis safeguards against cardiomyocyte apoptosis and cardiotoxicity induced by chemotherapy.
International journal of biological sciences
Liu C,Cheng X,Xing J,Li J,Li Z,Jian D,Wang Y,Wang S,Li R,Zhang W,Shao D,Ma X,Chen X,Shen J,Shi C,Guo Z,Wang W,Fan T,Liu L,Tang H
Thu May 12 00:00:00 EDT 2022
Connexin 43 (Cx43) regulates high-glucose-induced retinal endothelial cell angiogenesis and retinal neovascularization.
Frontiers in endocrinology
Shi W,Meng Z,Luo J
Tue Sep 20 00:00:00 EDT 2022
In Vitro Inhibitory Effects of Maqian Essential Oil against Ectopic Endometrial Stromal Cells and LPS-Induced Endometrial Epithelial Cells.
Chemistry & biodiversity
Li LP,Luo Y,Huang C,Wang XR,Huang TT,Zou YY,Huang SH,Liu YQ,Yang BC
Thu Dec 01 00:00:00 EST 2022
Bombyx mori Akirin hijacks a viral peptide vSP27 encoded by BmCPV circRNA and activates the ROS-NF-κB pathway against viral infection.
International journal of biological macromolecules
Zhang Y,Zhang X,Dai K,Zhu M,Liang Z,Pan J,Zhang Z,Xue R,Cao G,Hu X,Gong C
Sat Jan 01 00:00:00 EST 2022
Aging is associated with increased brain iron through cortex-derived hepcidin expression.
eLife
Sato T,Shapiro JS,Chang HC,Miller RA,Ardehali H
Tue Jan 11 00:00:00 EST 2022
Honokiol improves depression-like behaviors in rats by HIF-1α- VEGF signaling pathway activation.
Frontiers in pharmacology
Fan XX,Sun WY,Li Y,Tang Q,Li LN,Yu X,Wang SY,Fan AR,Xu XQ,Chang HS
Tue Sep 13 00:00:00 EDT 2022
M1 Macrophages Enhance Survival and Invasion of Oral Squamous Cell Carcinoma by Inducing GDF15-Mediated ErbB2 Phosphorylation.
ACS omega
Lv C,Li S,Zhao J,Yang P,Yang C
Tue Apr 05 00:00:00 EDT 2022
TMUB1 is an endoplasmic reticulum-resident escortase that promotes the p97-mediated extraction of membrane proteins for degradation.
Molecular cell
Wang L,Li J,Wang Q,Ge MX,Ji J,Liu D,Wang Z,Cao Y,Zhang Y,Zhang ZR
Thu Sep 15 00:00:00 EDT 2022
The central scaffold protein CEP350 coordinates centriole length, stability, and maturation.
The Journal of cell biology
Karasu OR,Neuner A,Atorino ES,Pereira G,Schiebel E
Mon Dec 05 00:00:00 EST 2022
LncRNA Riken Attenuated Sevoflurane-Induced Neuroinflammation by Regulating the MicroRNA-101a/MKP-1/JNK Pathway.
Neurotoxicity research
Hou Q,Li S,Zhang B,Chu H,Ni C,Fei X,Zheng H
Tue Feb 01 00:00:00 EST 2022
Lytic cocktail: An effective method to alleviate severe burn induced hyper-metabolism through regulating white adipose tissue browning.
Heliyon
Zhang M,Yang P,Yu T,Harmsen MC,Gao M,Liu D,Shi Y,Liu Y,Zhang X
Tue Mar 01 00:00:00 EST 2022
Estrogen-related Receptor α (ERRα) Functions in The Hypoxic Injury of Microglial Cells.
Journal of veterinary research
Deng CY,Zhu TT,Lian S,Wang JF,Wu R,Zheng JS
Tue Mar 01 00:00:00 EST 2022
TRIM25 inhibits influenza A virus infection, destabilizes viral mRNA, but is redundant for activating the RIG-I pathway.
Nucleic acids research
Choudhury NR,Trus I,Heikel G,Wolczyk M,Szymanski J,Bolembach A,Dos Santos Pinto RM,Smith N,Trubitsyna M,Gaunt E,Digard P,Michlewski G
Fri Jul 08 00:00:00 EDT 2022
High expression of aryl hydrocarbon receptor (AhR) plays an important role in the formation of fibrous epulis.
Oral diseases
Zeng X,Xia S,Yan X,Hu C,An K,Luo L
Tue Nov 01 00:00:00 EDT 2022
Anti-tumor effects and mechanism of a novel camptothecin derivative YCJ100.
Life sciences
Zhang M,Fu W,Zhu LZ,Liu XF,Li L,Peng LZ,Kai GY,Liu YQ,Zhang ZJ,Xu CR
Thu Dec 15 00:00:00 EST 2022
Nuclear Condensation of CDYL Links Histone Crotonylation and Cystogenesis in Autosomal Dominant Polycystic Kidney Disease.
Journal of the American Society of Nephrology : JASN
Dang L,Cao X,Zhang T,Sun Y,Tian S,Gong T,Xiong H,Cao P,Li Y,Yu S,Yang L,Zhang L,Liu T,Zhang K,Liang J,Chen Y
Thu Sep 01 00:00:00 EDT 2022
Activation of Piezo1 contributes to matrix stiffness-induced angiogenesis in hepatocellular carcinoma.
Cancer communications (London, England)
Li M,Zhang X,Wang M,Wang Y,Qian J,Xing X,Wang Z,You Y,Guo K,Chen J,Gao D,Zhao Y,Zhang L,Chen R,Cui J,Ren Z
Tue Nov 01 00:00:00 EDT 2022
The hsa-miR-214-3p/ATGL axis regulates aberrant lipolysis to promote acute myeloid leukemia progression via PPARα in vitro.
Biochemical and biophysical research communications
Li Y,Yuan S,Wu W,Zhou J,Zhang P,Li D,Zhang Y,Lou S
Sat Jun 11 00:00:00 EDT 2022
αTAT1-induced tubulin acetylation promotes ameloblastoma migration and invasion.
Laboratory investigation; a journal of technical methods and pathology
Yoshimoto S,Morita H,Okamura K,Hiraki A,Hashimoto S
Sat Jan 01 00:00:00 EST 2022
αTAT1-induced tubulin acetylation promotes ameloblastoma migration and invasion.
Laboratory investigation; a journal of technical methods and pathology
Yoshimoto S,Morita H,Okamura K,Hiraki A,Hashimoto S
Sat Jan 01 00:00:00 EST 2022
Engineering Lipusu with lysophosphatidylcholine for improved tumor cellular uptake and anticancer efficacy.
Journal of materials chemistry. B
Li L,Zhan Q,Yi K,Chen N,Li X,Yang S,Hou X,Zhao J,Yuan X,Kang C
Wed Mar 16 00:00:00 EDT 2022
The phytochemical, corynoline, diminishes Aurora kinase B activity to induce mitotic defect and polyploidy.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Yan Z,Shi Q,Liu X,Li J,Ahire V,Zhang S,Zhang J,Yang D,Allen TD
Tue Mar 01 00:00:00 EST 2022
Intestinal Epithelial STAT6 Activation Rescues the Defective Anti-Helminth Responses Caused by Ogt Deletion.
International journal of molecular sciences
Xiong X,Huang R,Li Z,Yang C,Wang Q,Ruan HB,Xu L
Thu Sep 22 00:00:00 EDT 2022
Theaflavin-3,3'-Digallate from Black Tea Inhibits Neointima Formation Through Suppression of the PDGFRβ Pathway in Vascular Smooth Muscle Cells.
Frontiers in pharmacology
Wu Y,Chen M,Chen Z,Shu J,Zhang L,Hu J,Yu H,Huang K,Liang M
Sun Jul 31 00:00:00 EDT 2022
Involvement of Rab11 in osteoblastic differentiation: Its up-regulation during the differentiation and by tensile stress.
Biochemical and biophysical research communications
Thant L,Kakihara Y,Kaku M,Kitami M,Kitami K,Mizukoshi M,Maeda T,Saito I,Saeki M
Sat Oct 08 00:00:00 EDT 2022
Carbon Monoxide-Releasing Molecule-3 Enhances Osteogenic Differentiation of Rat Bone Marrow Mesenchymal Stem Cells via miR-195-5p/Wnt3a Pathway.
Drug design, development and therapy
Li J,Han Q,Chen H,Liu T,Song J,Hou M,Wei L,Song H
Tue Jul 12 00:00:00 EDT 2022
Avermectin induces carp neurotoxicity by mediating blood-brain barrier dysfunction, oxidative stress, inflammation, and apoptosis through PI3K/Akt and NF-κB pathways.
Ecotoxicology and environmental safety
Zhang T,Dong Z,Liu F,Pan E,He N,Ma F,Wang G,Wang Y,Dong J
Thu Sep 15 00:00:00 EDT 2022
Capsaicin suppresses hepatocarcinogenesis by inhibiting the stemness of hepatic progenitor cells via SIRT1/SOX2 signaling pathway.
Cancer medicine
Xie ZQ,Li HX,Hou XJ,Huang MY,Zhu ZM,Wei LX,Tang CX
Tue Nov 01 00:00:00 EDT 2022
Macrophage derived miR-7219-3p-containing exosomes mediate fibroblast trans-differentiation by targeting SPRY1 in silicosis.
Toxicology
Niu Z,Wang L,Qin X,Ye Z,Xie B,Hu Y
Thu Sep 01 00:00:00 EDT 2022
Identification of purine biosynthesis as an NADH-sensing pathway to mediate energy stress.
Nature communications
Yang R,Yang C,Ma L,Zhao Y,Guo Z,Niu J,Chu Q,Ma Y,Li B
Thu Nov 17 00:00:00 EST 2022
RNF31 represses cell progression and immune evasion via YAP/PD-L1 suppression in triple negative breast Cancer.
Journal of experimental & clinical cancer research : CR
Yang H,Xue M,Su P,Zhou Y,Li X,Li Z,Xia Y,Zhang C,Fu M,Zheng X,Luo G,Wei T,Wang X,Ding Y,Zhu J,Zhuang T
Thu Dec 29 00:00:00 EST 2022
The transcription factor KLF14 regulates macrophage glycolysis and immune function by inhibiting HK2 in sepsis.
Cellular & molecular immunology
Yuan Y,Fan G,Liu Y,Liu L,Zhang T,Liu P,Tu Q,Zhang X,Luo S,Yao L,Chen F,Li J
Fri Apr 01 00:00:00 EDT 2022
PINK1-parkin-mediated neuronal mitophagy deficiency in prion disease.
Cell death & disease
Li J,Lai M,Zhang X,Li Z,Yang D,Zhao M,Wang D,Sun Z,Ehsan S,Li W,Gao H,Zhao D,Yang L
Fri Feb 18 00:00:00 EST 2022
IRF2-ferroptosis related gene is associated with prognosis and EMT in gliomas.
Translational oncology
Tong S,Ye L,Xu Y,Sun Q,Gao L,Cai J,Ye Z,Tian D,Chen Q
Thu Dec 01 00:00:00 EST 2022
A mechanosensitive lipolytic factor in the bone marrow promotes osteogenesis and lymphopoiesis.
Cell metabolism
Peng H,Hu B,Xie LQ,Su T,Li CJ,Liu Y,Yang M,Xiao Y,Feng X,Zhou R,Guo Q,Zhou HY,Huang Y,Jiang TJ,Luo XH
Tue Aug 02 00:00:00 EDT 2022
Effects of Artemisinin on Escherichia coli-Induced Mastitis in Bovine Mammary Epithelial Cells and Mice.
Veterinary sciences
Li Z,Hu J,Wang X,Du Y,Yin J,Gao J,Han B,Cui S,Liu Y,Liu J
Tue Jul 26 00:00:00 EDT 2022
The Therapeutic Effect and the Possible Mechanism of C-Phycocyanin in Lipopolysaccharide and Seawater-Induced Acute Lung Injury.
Drug design, development and therapy
Zhang L,Kong D,Huang J,Wang Q,Shao L
Fri Apr 15 00:00:00 EDT 2022
Dehydrocostus Lactone Suppresses Dextran Sulfate Sodium-Induced Colitis by Targeting the IKKα/β-NF-κB and Keap1-Nrf2 Signalling Pathways.
Frontiers in pharmacology
Yuan Y,Hu Q,Liu L,Xie F,Yang L,Li Y,Zhang C,Chen H,Tang J,Shen X
Fri Mar 25 00:00:00 EDT 2022
Effect of nicotinamide N-methyltransferase on lipid accumulation in 3T3-L1 adipocytes.
Bioengineered
Xu W,Hou L,Li P,Li L
Sun May 01 00:00:00 EDT 2022
Suppression of ACE2 SUMOylation protects against SARS-CoV-2 infection through TOLLIP-mediated selective autophagy.
Nature communications
Jin S,He X,Ma L,Zhuang Z,Wang Y,Lin M,Cai S,Wei L,Wang Z,Zhao Z,Wu Y,Sun L,Li C,Xie W,Zhao Y,Songyang Z,Peng K,Zhao J,Cui J
Sat Sep 03 00:00:00 EDT 2022
Inhibition of translocator protein 18 kDa suppressed the progression of glioma via the ELAV-like RNA-binding protein 1/MAPK-activated protein kinase 3 axis.
Bioengineered
Wang J,Ren P,Zeng Z,Ma L,Li Y,Zhang H,Guo W
Tue Mar 01 00:00:00 EST 2022
Therapeutic targeting of glutamate dehydrogenase 1 that links metabolic reprogramming and Snail-mediated epithelial-mesenchymal transition in drug-resistant lung cancer.
Pharmacological research
Wang Q,Wu M,Li H,Rao X,Ao L,Wang H,Yao L,Wang X,Hong X,Wang J,Aa J,Sun M,Wang G,Liu J,Zhou F
Tue Nov 01 00:00:00 EDT 2022
METTL3-mediated m6A RNA methylation regulates dorsal lingual epithelium homeostasis.
International journal of oral science
Xiong Q,Liu C,Zheng X,Zhou X,Lei K,Zhang X,Wang Q,Lin W,Tong R,Xu R,Yuan Q
Tue May 17 00:00:00 EDT 2022
Virus-induced FoxO factor facilitates replication of human cytomegalovirus.
Archives of virology
Sleman S
Sat Jan 01 00:00:00 EST 2022
Effect of resistance training on satellite cells in old mice - a transcriptome study : implications for sarcopenia.
Bone & joint research
Hsu WB,Lin SJ,Hung JS,Chen MH,Lin CY,Hsu WH,Hsu WR
Tue Feb 01 00:00:00 EST 2022
The TLR-chaperone CNPY3 is a critical regulator of NLRP3-inflammasome activation.
European journal of immunology
Ghait M,Husain RA,Duduskar SN,Haack TB,Rooney M,Göhrig B,Bauer M,Rubio I,Deshmukh SD
Wed Jun 01 00:00:00 EDT 2022
Successful outcomes of intracytoplasmic sperm injection-embryo transfer using ejaculated spermatozoa from two Chinese asthenoteratozoospermic brothers with a compound heterozygous FSIP2 mutation.
Andrologia
Yuan Y,Xu WQ,Chen ZY,Chen Y,Zhang L,Zheng L,Luo T,Chen HY
Fri Apr 01 00:00:00 EDT 2022
EspF of Enterohemorrhagic Escherichia coli Enhances Apoptosis via Endoplasmic Reticulum Stress in Intestinal Epithelial Cells: An Isobaric Tags for Relative and Absolute Quantitation-Based Comparative Proteomic Analysis.
Frontiers in microbiology
Wang X,Yan K,Fu M,Liang S,Zhao H,Fu C,Yang L,Song Z,Sun D,Wan C
Tue Jul 19 00:00:00 EDT 2022
Regulation of TET2 gene expression and 5mC oxidation in breast cancer cells by estrogen signaling.
Biochemical and biophysical research communications
Zhu X,Xiong L,Lyu R,Shen Y,Liu L,Li S,Argueta C,Tan L
Sat Jan 22 00:00:00 EST 2022
PRSS55 is a novel potential causative gene for human male infertility.
Reproductive biomedicine online
Liu M,Jiang C,Zhang X,Zhang G,Liu M,Zheng R,Yang Y,Shen Y
Thu Sep 01 00:00:00 EDT 2022
PRSS55 is a novel potential causative gene for human male infertility.
Reproductive biomedicine online
Liu M,Jiang C,Zhang X,Zhang G,Liu M,Zheng R,Yang Y,Shen Y
Thu Sep 01 00:00:00 EDT 2022
Fumarate inhibits PTEN to promote tumorigenesis and therapeutic resistance of type2 papillary renal cell carcinoma.
Molecular cell
Ge X,Li M,Yin J,Shi Z,Fu Y,Zhao N,Chen H,Meng L,Li X,Hu Z,Zhao X,Guo H,Qian X
Thu Apr 07 00:00:00 EDT 2022
Non-canonical Small GTPase RBJ Promotes NSCLC Progression Through the Canonical MEK/ERK Signaling Pathway.
Current pharmaceutical design
Wang Y,Shen X,Wang Q,Guo Z,Hu L,Dong Z,Hu W
Fri Jan 06 00:00:00 EST 2023
Curcumin reduces oxidative stress and fat deposition in longissimus dorsi muscle of intrauterine growth-retarded finishing pigs.
Animal science journal = Nihon chikusan Gakkaiho
Zhang J,Yan E,Zhang L,Wang T,Wang C
Fri Jun 17 00:00:00 EDT 2022
WNT6 participates in the occurrence and development of ovarian cancer by upregulating/activating the typical Wnt pathway and Notch1 signaling pathway.
Gene
Bao H,Wu W,Li Y,Zong Z,Chen S
Tue Dec 20 00:00:00 EST 2022
The Acetylation Modification of SP1 Regulates the Protein Stability in Silkworm.
Applied biochemistry and biotechnology
Sun Z,Ma Y,Liu Y,Lv J,Wang D,You Z,Jiang C,Sheng Q,Nie Z
Fri Apr 01 00:00:00 EDT 2022
Immune Characteristics and Prognosis Analysis of the Proteasome 20S Subunit Beta 9 in Lower-Grade Gliomas.
Frontiers in oncology
Liu J,Yang X,Ji Q,Yang L,Li J,Long X,Ye M,Huang K,Zhu X
Sat Aug 06 00:00:00 EDT 2022
GPR108 is required for gambogic acid inhibiting NF-κB signaling in cancer.
Pharmacological research
Lyu S,Zhang X,Tu Z,Zhou H,Ke X,Qu Y
Mon Aug 01 00:00:00 EDT 2022
The Pathogenic Mechanism of the ATP2C1 p.Ala109_Gln120del Mutation in Hailey-Hailey Disease.
Clinical, cosmetic and investigational dermatology
Li P,Qi J,Zhou B,Ding T,Long J,Xiao H
Tue Nov 01 00:00:00 EDT 2022
Inka2, a novel Pak4 inhibitor, regulates actin dynamics in neuronal development.
PLoS genetics
Yamada S,Mizukoshi T,Tokunaga A,Sakakibara SI
Sat Oct 01 00:00:00 EDT 2022
Inka2, a novel Pak4 inhibitor, regulates actin dynamics in neuronal development.
PLoS genetics
Yamada S,Mizukoshi T,Tokunaga A,Sakakibara SI
Sat Oct 01 00:00:00 EDT 2022
Dihydroartemisinin enhances the inhibitory effect of sorafenib on HepG2 cells by inducing ferroptosis and inhibiting energy metabolism.
Journal of pharmacological sciences
Cui Z,Wang H,Li S,Qin T,Shi H,Ma J,Li L,Yu G,Jiang T,Li C
Sat Jan 01 00:00:00 EST 2022
Hydrogen-Rich Medium Regulates Cr(VI)-Induced ER Stress and Autophagy Signaling in DF-1 Cells.
Biological trace element research
Liu K,Cui Y,Li H,Qi C,Cheng G,Gao X,Zhang Z,Liu Y,Liu J
Sun May 01 00:00:00 EDT 2022
SFN Enhanced the Radiosensitivity of Cervical Cancer Cells via Activating LATS2 and Blocking Rad51/MDC1 Recruitment to DNA Damage Site.
Cancers
Wang S,Wang Y,Liu X,Yang Y,Wu S,Liu Y
Fri Apr 08 00:00:00 EDT 2022
HECTD3 promotes gastric cancer progression by mediating the polyubiquitination of c-MYC.
Cell death discovery
Zhang G,Zhu Q,Yan X,Ci M,Zhao E,Hou J,Wan S,Lü M,Cui H
Sat Apr 09 00:00:00 EDT 2022
The ethanol extract of flower buds of Tussilago farfara L. attenuates cigarette smoke-induced lung inflammation through regulating NLRP3 inflammasome, Nrf2, and NF-κB.
Journal of ethnopharmacology
Xu LT,Wang T,Fang KL,Zhao Y,Wang XN,Ren DM,Shen T
Sun Jan 30 00:00:00 EST 2022
Fine-Tuning of Cholesterol Homeostasis Controls Erythroid Differentiation.
Advanced science (Weinheim, Baden-Wurttemberg, Germany)
Lu Z,Huang L,Li Y,Xu Y,Zhang R,Zhou Q,Sun Q,Lu Y,Chen J,Shen Y,Li J,Zhao B
Sat Jan 01 00:00:00 EST 2022
Morusin enhances the antitumor activity of MAPK pathway inhibitors in BRAF-mutant melanoma by inhibiting the feedback activation of STAT3.
European journal of cancer (Oxford, England : 1990)
Zhao K,Dai Q,Wu J,Wei Z,Duan Y,Chen B
Fri Apr 01 00:00:00 EDT 2022
C-terminal glutamine acts as a C-degron targeted by E3 ubiquitin ligase TRIM7.
Proceedings of the National Academy of Sciences of the United States of America
Ru Y,Yan X,Zhang B,Song L,Feng Q,Ye C,Zhou Z,Yang Z,Li Y,Zhang Z,Li Q,Mi W,Dong C
Tue Jul 26 00:00:00 EDT 2022
SYTL5 Promotes Papillary Thyroid Carcinoma Progression by Enhancing Activation of the NF-κB Signaling Pathway.
Endocrinology
Huang D,Zeng Y,Deng HY,Fu BD,Ke Y,Luo JY,Yu JC,Yu ST
Mon Nov 14 00:00:00 EST 2022
ZEB2/TWIST1/PRMT5/NuRD Multicomplex Contributes to the Epigenetic Regulation of EMT and Metastasis in Colorectal Carcinoma.
Cancers
Zheng Y,Dai M,Dong Y,Yu H,Liu T,Feng X,Yu B,Zhang H,Wu J,Kong W,Yu X,Wu H
Thu Jul 14 00:00:00 EDT 2022
The interaction between E3 ubiquitin ligase Parkin and mitophagy receptor PHB2 links inner mitochondrial membrane ubiquitination to efficient mitophagy.
The Journal of biological chemistry
Sun S,Hou H,Ma G,Ma Q,Li N,Zhang L,Dong C,Cao M,Tam KY,Ying Z,Wang H
Thu Dec 01 00:00:00 EST 2022
Luteolin attenuated cisplatin-induced cardiac dysfunction and oxidative stress via modulation of Keap1/Nrf2 signaling pathway.
Free radical research
Qi Y,Fu S,Pei D,Fang Q,Xin W,Yuan X,Cao Y,Shu Q,Mi X,Luo F
Tue Feb 01 00:00:00 EST 2022
cAMP-Induced Nuclear Condensation of CRTC2 Promotes Transcription Elongation and Cystogenesis in Autosomal Dominant Polycystic Kidney Disease.
Advanced science (Weinheim, Baden-Wurttemberg, Germany)
Mi Z,Song Y,Wang J,Liu Z,Cao X,Dang L,Lu Y,Sun Y,Xiong H,Zhang L,Chen Y
Fri Apr 01 00:00:00 EDT 2022
Tubastatin A Improves Post-Resuscitation Myocardial Dysfunction by Inhibiting NLRP3-Mediated Pyroptosis Through Enhancing Transcription Factor EB Signaling.
Journal of the American Heart Association
Xu J,Zhao X,Jiang X,He L,Wu X,Wang J,Chen Q,Li Y,Zhang M
Tue Apr 05 00:00:00 EDT 2022
Reversal of Lipid Metabolism Dysregulation by Selenium and Folic Acid Co-Supplementation to Mitigate Pathology in Alzheimer's Disease.
Antioxidants (Basel, Switzerland)
Zhang ZH,Cao XC,Peng JY,Huang SL,Chen C,Jia SZ,Ni JZ,Song GL
Sun Apr 24 00:00:00 EDT 2022
Hyaluronic Acid Facilitates Angiogenesis of Endothelial Colony Forming Cell Combining With Mesenchymal Stem Cell via CD44/ MicroRNA-139-5p Pathway.
Frontiers in bioengineering and biotechnology
Luo Y,Liang F,Wan X,Liu S,Fu L,Mo J,Meng X,Mo Z
Fri Apr 01 00:00:00 EDT 2022
Targeting HECTD3-IKKα axis inhibits inflammation-related metastasis.
Signal transduction and targeted therapy
Li F,Liang H,You H,Xiao J,Xia H,Chen X,Huang M,Cheng Z,Yang C,Liu W,Zhang H,Zeng L,Wu Y,Ge F,Li Z,Zhou W,Wen Y,Zhou Z,Liu R,Jiang D,Xie N,Liang B,Liu Z,Kong Y,Chen C
Wed Aug 03 00:00:00 EDT 2022
Iron-regulated assembly of the cytosolic iron-sulfur cluster biogenesis machinery.
The Journal of biological chemistry
Fan X,Barshop WD,Vashisht AA,Pandey V,Leal S,Rayatpisheh S,Jami-Alahmadi Y,Sha J,Wohlschlegel JA
Fri Jul 01 00:00:00 EDT 2022
Planar cell polarity induces local microtubule bundling for coordinated ciliary beating.
The Journal of cell biology
Nakayama S,Yano T,Namba T,Konishi S,Takagishi M,Herawati E,Nishida T,Imoto Y,Ishihara S,Takahashi M,Furuta K,Oiwa K,Tamura A,Tsukita S
Mon Jul 05 00:00:00 EDT 2021
Planar cell polarity induces local microtubule bundling for coordinated ciliary beating.
The Journal of cell biology
Nakayama S,Yano T,Namba T,Konishi S,Takagishi M,Herawati E,Nishida T,Imoto Y,Ishihara S,Takahashi M,Furuta K,Oiwa K,Tamura A,Tsukita S
Mon Jul 05 00:00:00 EDT 2021
WDR62 regulates spindle dynamics as an adaptor protein between TPX2/Aurora A and katanin.
The Journal of cell biology
Huang J,Liang Z,Guan C,Hua S,Jiang K
Mon Aug 02 00:00:00 EDT 2021
WDR62 regulates spindle dynamics as an adaptor protein between TPX2/Aurora A and katanin.
The Journal of cell biology
Huang J,Liang Z,Guan C,Hua S,Jiang K
Mon Aug 02 00:00:00 EDT 2021
DMRT2 Interacts With FXR and Improves Insulin Resistance in Adipocytes and a Mouse Model.
Frontiers in endocrinology
Tao J,Yu XL,Yuan YJ,Shen X,Liu J,Gu PP,Wang Z,Ma YT,Li GQ
Fri Apr 22 00:00:00 EDT 2022
Mefunidone ameliorates diabetic kidney disease in STZ and db/db mice.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Jiang Y,Xie F,Lv X,Wang S,Liao X,Yu Y,Dai Q,Zhang Y,Meng J,Hu G,Peng Z,Tao L
Fri Jan 01 00:00:00 EST 2021
MiR-155-5p suppresses SOX1 to promote proliferation of cholangiocarcinoma via RAF/MEK/ERK pathway.
Cancer cell international
Wang D,Xiong F,Wu G,Liu W,Wang B,Chen Y
Tue Dec 07 00:00:00 EST 2021
Exosomes derived from thymic stromal lymphopoietin-treated dendritic cells regulate T helper 17/regulatory T cell differentiation via miR-21/Smad7 axis.
Experimental cell research
Ji Q,Liu J,Dong Y,Wang L,Dong K,Setiz B,Szentmáry N,Qu B,Shi L
Fri Jan 01 00:00:00 EST 2021
Deacetylation of BmAda3 is required for cell apoptosis caused by Bombyx mori nucleopolyhedrovirus infection.
Archives of insect biochemistry and physiology
Zhu Y,Hu M,Ngowo J,Gao X,Chen X,Yan H,Yu W
Fri Oct 01 00:00:00 EDT 2021
Salmonella Effector SpvB Inhibits NF-κB Activity via KEAP1-Mediated Downregulation of IKKβ.
Frontiers in cellular and infection microbiology
Yang S,Deng Q,Sun L,Zhu Y,Dong K,Wu S,Huang R,Li Y
Mon Jul 05 00:00:00 EDT 2021
CSN6 promotes melanoma proliferation and metastasis by controlling the UBR5-mediated ubiquitination and degradation of CDK9.
Cell death & disease
Zhang Y,Hou J,Shi S,Du J,Liu Y,Huang P,Li Q,Liu L,Hu H,Ji Y,Guo L,Shi Y,Liu Y,Cui H
Fri Jan 22 00:00:00 EST 2021
Oleic Acid and Insulin as Key Characteristics of T2D Promote Colorectal Cancer Deterioration in Xenograft Mice Revealed by Functional Metabolomics.
Frontiers in oncology
Zhang Y,Wang D,Lv B,Hou X,Liu Q,Liao C,Xu R,Zhang Y,Xu F,Zhang P
Fri Aug 27 00:00:00 EDT 2021
Effects and Mechanisms of Synaptotagmin-7 in the Hippocampus on Cognitive Impairment in Aging Mice.
Molecular neurobiology
Xie Y,Zhi K,Meng X
Mon Nov 01 00:00:00 EDT 2021
Design, Synthesis, and Biological Evaluation of Icaritin Derivatives as Novel Putative DEPTOR Inhibitors for Multiple Myeloma Treatment.
Journal of medicinal chemistry
Hou Y,Kuang W,Min W,Liu Z,Zhang F,Yuan K,Wang X,Sun C,Cheng H,Wang L,Xiao Y,Pu S,Xin GZ,Yang P
Thu Oct 28 00:00:00 EDT 2021
Pharmacological Inhibition of HMGB1 Prevents Muscle Wasting.
Frontiers in pharmacology
Li L,Liu H,Tao W,Wen S,Fu X,Yu S
Tue Dec 07 00:00:00 EST 2021
PFKP Activation Ameliorates Foot Process Fusion in Podocytes in Diabetic Kidney Disease.
Frontiers in endocrinology
Zhang Z,Liang W,Luo Q,Hu H,Yang K,Hu J,Chen Z,Zhu J,Feng J,Zhu Z,Chi Q,Ding G
Wed Mar 02 00:00:00 EST 2022
Comprehensive Bioinformatics Analysis Identifies POLR2I as a Key Gene in the Pathogenesis of Hypertensive Nephropathy.
Frontiers in genetics
You S,Xu J,Wu B,Wu S,Zhang Y,Sun Y,Zhang N
Tue Aug 24 00:00:00 EDT 2021
The antitumor role of a newly discovered α-d-glucan from Holotrichia diomphalia Bates as a selective blocker of aldolase A.
Carbohydrate polymers
Wang J,Li Z,Yang X,Qiao Y,Feng C,Yu S,Jing H,Liu W,Ren L,Duan Q,Li XQ,Cao W
Mon Mar 01 00:00:00 EST 2021
WIPI-1 inhibits metastasis and tumour growth via the WIPI-1-TRIM21 axis and MYC regulation in nasopharyngeal carcinoma.
Oral oncology
Zhao Y,Li WF,Li QJ,He SW,He QM,Long LF,Liu N,Ma J
Mon Nov 01 00:00:00 EDT 2021
Keratin 19 maintains E-cadherin localization at the cell surface and stabilizes cell-cell adhesion of MCF7 cells.
Cell adhesion & migration
Alsharif S,Sharma P,Bursch K,Milliken R,Lam V,Fallatah A,Phan T,Collins M,Dohlman P,Tiufekchiev S,Nehmetallah G,Raub CB,Chung BM
Wed Dec 01 00:00:00 EST 2021
RNF220-mediated ubiquitination promotes aggresomal accumulation and autophagic degradation of cytoplasmic Gli via HDAC6.
Biochemical and biophysical research communications
Wang H,Yang C,Li Y,Zhao S,Ma P,Mao B
Fri Jun 11 00:00:00 EDT 2021
Iron deficiency promotes aortic medial degeneration via destructing cytoskeleton of vascular smooth muscle cells.
Clinical and translational medicine
Li B,Wang Z,Hong J,Che Y,Chen R,Hu Z,Hu X,Wu Q,Hu J,Zhang M
Fri Jan 01 00:00:00 EST 2021
Genistein suppresses allergic contact dermatitis through regulating the MAP2K2/ERK pathway.
Food & function
Xu J,Xiong H,Zhao Z,Luo M,Ju Y,Yang G,Mei Z
Fri May 21 00:00:00 EDT 2021
Aberrantly high activation of a FoxM1-STMN1 axis contributes to progression and tumorigenesis in FoxM1-driven cancers.
Signal transduction and targeted therapy
Liu J,Li J,Wang K,Liu H,Sun J,Zhao X,Yu Y,Qiao Y,Wu Y,Zhang X,Zhang R,Yang A
Mon Feb 01 00:00:00 EST 2021
Modulated TRPC1 Expression Predicts Sensitivity of Breast Cancer to Doxorubicin and Magnetic Field Therapy: Segue Towards a Precision Medicine Approach.
Frontiers in oncology
Tai YK,Chan KKW,Fong CHH,Ramanan S,Yap JLY,Yin JN,Yip YS,Tan WR,Koh APF,Tan NS,Chan CW,Huang RYJ,Li JZ,Fröhlich J,Franco-Obregón A
Sat Jul 16 00:00:00 EDT 2022
C23 gene regulates the nucleolin structure and biosynthesis of ribosomes in bovine intraspecific and interspecific somatic cell nuclear transfer embryos.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Su G,Wang L,Gao G,Wu S,Yang L,Wu M,Liu X,Yang M,Wei Z,Bai C,Li G
Mon Nov 01 00:00:00 EDT 2021
VPS13D interacts with VCP/p97 and negatively regulates endoplasmic reticulum-mitochondria interactions.
Molecular biology of the cell
Du Y,Wang J,Xiong J,Fang N,Ji WK
Sun Aug 01 00:00:00 EDT 2021
Small molecule targeting topoisomerase 3β for cancer therapy.
Pharmacological research
Zhang X,Wang L,Zhang Q,Lyu S,Zhu D,Shen M,Ke X,Qu Y
Wed Dec 01 00:00:00 EST 2021
Dihydrocapsaicin Inhibits Cell Proliferation and Metastasis in Melanoma via Down-regulating β-Catenin Pathway.
Frontiers in oncology
Shi S,Li C,Zhang Y,Deng C,Liu W,Du J,Li Q,Ji Y,Guo L,Liu L,Hu H,Liu Y,Cui H
Sat Nov 11 00:00:00 EST 2023
Pan-Cancer Analyses Reveal Oncogenic Role and Prognostic Value of F-Box Only Protein 22.
Frontiers in oncology
Chen S,Ma S,Yan J,Wang H,Ding B,Guo Z,Ma Y,Chen X,Wang Y
Wed Feb 16 00:00:00 EST 2022
Generation of VEGF knock-in Cashmere goat via the CRISPR/Cas9 system.
International journal of biological sciences
Hu X,Hao F,Li X,Xun Z,Gao Y,Ren B,Cang M,Liang H,Liu D
Fri Jan 21 00:00:00 EST 2022
Aagab acts as a novel regulator of NEDD4-1-mediated Pten nuclear translocation to promote neurological recovery following hypoxic-ischemic brain damage.
Cell death and differentiation
Dai C,Wu B,Chen Y,Li X,Bai Y,Du Y,Pang Y,Wang YT,Dong Z
Sun Aug 01 00:00:00 EDT 2021
ZFC3H1 prevents RNA trafficking into nuclear speckles through condensation.
Nucleic acids research
Wang Y,Fan J,Wang J,Zhu Y,Xu L,Tong D,Cheng H
Mon Oct 11 00:00:00 EDT 2021
Initiation of oogenesis and meiosis in the fetal ovary depends on Dennd1a-mediated production of Wnt5a and retinoic acid from the somatic niches.
Frontiers in bioscience (Landmark edition)
Shi J,Niu Q,Gao Q,Fu J,Ma J
Thu Dec 30 00:00:00 EST 2021
PGK1 Is a Key Target for Anti-Glycolytic Therapy of Ovarian Cancer: Based on the Comprehensive Analysis of Glycolysis-Related Genes.
Frontiers in oncology
Gou R,Hu Y,Liu O,Dong H,Gao L,Wang S,Zheng M,Li X,Lin B
Tue Jul 20 00:00:00 EDT 2021
Neuroprotective effects of kukoamine A on 6-OHDA-induced Parkinson's model through apoptosis and iron accumulation inhibition.
Chinese herbal medicines
Li X,Jiang XW,Chu HX,Zhao QC,Ding HW,Cai CH
Fri Jan 01 00:00:00 EST 2021
Cordycepin inhibits the proliferation of malignant peripheral nerve sheath tumor cells through the p53/Sp1/tubulin pathway.
American journal of cancer research
Lee MJ,Lee JC,Hsieh JH,Lin MY,Shih IA,You HL,Wang K
Fri May 07 00:00:00 EDT 2021
Histone Deacetylase Inhibitor SAHA Induces Expression of Fatty Acid-Binding Protein 4 and Inhibits Replication of Human Cytomegalovirus.
Virologica Sinica
Liu Z,Xuan B,Tang S,Qian Z
Wed Dec 01 00:00:00 EST 2021
Periodontal Pathogens Promote Oral Squamous Cell Carcinoma by Regulating ATR and NLRP3 Inflammasome.
Frontiers in oncology
Yao Y,Shen X,Zhou M,Tang B
Fri Oct 22 00:00:00 EDT 2021
Bardoxolone-Methyl Prevents Oxidative Stress-Mediated Apoptosis and Extracellular Matrix Degradation in vitro and Alleviates Osteoarthritis in vivo.
Drug design, development and therapy
Pang Z,Jiang Z,Zhu R,Song C,Tang H,Cao L,Guo C
Thu Jan 20 00:00:00 EST 2022
The effects of myeloablative or non-myeloablative total body irradiations on intestinal tract in mice.
Bioscience reports
Zhu S,Liang J,Zhu F,Zhang X,Xu M,Zhao K,Zeng L,Xu K
Fri Mar 26 00:00:00 EDT 2021
DNA methylation mediated RSPO2 to promote follicular development in mammals.
Cell death & disease
Zhou X,He Y,Li N,Bai G,Pan X,Zhang Z,Zhang H,Li J,Yuan X
Sat Jun 26 00:00:00 EDT 2021
Silencing ZIC2 abrogates tumorigenesis and anoikis resistance of non-small cell lung cancer cells by inhibiting Src/FAK signaling.
Molecular therapy oncolytics
Liu A,Xie H,Li R,Ren L,Yang B,Dai L,Lu W,Liu B,Ren D,Zhang X,Chen Q,Huang Y,Shi K
Fri Sep 24 00:00:00 EDT 2021
Loss-of-function missense variant of AKAP4 induced male infertility through reduced interaction with QRICH2 during sperm flagella development.
Human molecular genetics
Zhang G,Li D,Tu C,Meng L,Tan Y,Ji Z,Cheng J,Lu G,Lin G,Zhang H,Sun J,Wang M,Du J,Xu W
Mon Dec 27 00:00:00 EST 2021
ZNRF2 attenuates focal cerebral ischemia/reperfusion injury in rats by inhibiting mTORC1-mediated autophagy.
Experimental neurology
Gu C,Yang J,Luo Y,Ran D,Tan X,Xiang P,Fei H,Lu Y,Guo W,Tu Y,Liu X,Wang H
Sun Aug 01 00:00:00 EDT 2021
Model-based analysis uncovers mutations altering autophagy selectivity in human cancer.
Nature communications
Han Z,Zhang W,Ning W,Wang C,Deng W,Li Z,Shang Z,Shen X,Liu X,Baba O,Morita T,Chen L,Xue Y,Jia D
Mon May 31 00:00:00 EDT 2021
Skip participates in formation and lipid metabolism of beige adipose and might mediate the effects of SIRT1 activator BTM-0512 on beige remodeling.
Biochemical and biophysical research communications
Liao ZM,Li AN,Cai Y,Chen JJ,Xu Y,Sui LH,Wang JL,Jin P,Wang KS,Yang ZC
Fri Jan 22 00:00:00 EST 2021
Loss of prion protein control of glucose metabolism promotes neurodegeneration in model of prion diseases.
PLoS pathogens
Arnould H,Baudouin V,Baudry A,Ribeiro LW,Ardila-Osorio H,Pietri M,Caradeuc C,Soultawi C,Williams D,Alvarez M,Crozet C,Djouadi F,Laforge M,Bertho G,Kellermann O,Launay JM,Schmitt-Ulms G,Schneider B
Fri Oct 01 00:00:00 EDT 2021
Aiduqing formula inhibits breast cancer metastasis by suppressing TAM/CXCL1-induced Treg differentiation and infiltration.
Cell communication and signaling : CCS
Li J,Wang S,Wang N,Zheng Y,Yang B,Wang X,Zhang J,Pan B,Wang Z
Mon Aug 30 00:00:00 EDT 2021
Potential Role of SWI/SNF Complex Subunit Actin-Like Protein 6A in Cervical Cancer.
Frontiers in oncology
Wang Q,Cao Z,Wei Y,Zhang J,Cheng Z
Tue Aug 17 00:00:00 EDT 2021
CBL aggravates Ang II-induced cardiac hypertrophy via the VHL/HIF-1α pathway.
Experimental cell research
Yang Y,Zou P,He L,Shao J,Tang Y,Li J
Sun Aug 15 00:00:00 EDT 2021
Actin-granule formation is an additional step in cardiac myofibroblast differentiation.
Annals of translational medicine
He L,Liu R,Yue H,Ren S,Zhu G,Guo Y,Qin C
Fri Jan 01 00:00:00 EST 2021
A human β-casein-derived peptide BCCY-1 modulates the innate immune response.
Food chemistry
Cai J,Li X,Wang X,Jiang C,Shen D,Cui X,Xie K,Ji C,Cao Y
Wed Jun 30 00:00:00 EDT 2021
Engorgement of Rhipicephalus haemaphysaloides ticks blocked by silencing a protein inhibitor of apoptosis.
Experimental & applied acarology
Tuerdi M,Hu S,Wang Y,Zhou Y,Cao J,Zhang H,Zhou J
Thu Jul 01 00:00:00 EDT 2021
Trojan Horse Delivery of 4,4'-Dimethoxychalcone for Parkinsonian Neuroprotection.
Advanced science (Weinheim, Baden-Wurttemberg, Germany)
Zhang W,Chen H,Ding L,Gong J,Zhang M,Guo W,Xu P,Li S,Zhang Y
Sat May 01 00:00:00 EDT 2021
A-FABP-PTEN/AKT Regulates Insulin Resistance in Preadipocyte Cell 3T3-L1 Cells.
Diabetes, metabolic syndrome and obesity : targets and therapy
Wu R,Wang H,Huangfu J,Xiao R
Sat Apr 23 00:00:00 EDT 2022
Protective effects of total flavonoids from Qu Zhi Qiao (fruit of Citrus paradisi cv. Changshanhuyou) on OVA-induced allergic airway inflammation and remodeling through MAPKs and Smad2/3 signaling pathway.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Wang J,Li T,Cai H,Jin L,Li R,Shan L,Cai W,Jiang J
Tue Jun 01 00:00:00 EDT 2021
Identification METTL18 as a Potential Prognosis Biomarker and Associated With Immune Infiltrates in Hepatocellular Carcinoma.
Frontiers in oncology
Li TH,Qin C,Zhao BB,Cao HT,Yang XY,Wang YY,Li ZR,Zhou XT,Wang WB
Tue Jun 15 00:00:00 EDT 2021
Vezatin regulates seizures by controlling AMPAR-mediated synaptic activity.
Cell death & disease
Wang Y,Yuan J,Yu X,Liu X,Tan C,Chen Y,Xu T
Tue Oct 12 00:00:00 EDT 2021
Effects of cigarette smoke on the aggravation of ovalbumin-induced asthma and the expressions of TRPA1 and tight junctions in mice.
Molecular immunology
Sun YB,Liu M,Fan XS,Zhou LP,Li MW,Hu FY,Yue QF,Zhang YM
Thu Jul 01 00:00:00 EDT 2021
HO-1 nuclear accumulation and interaction with NPM1 protect against stress-induced endothelial senescence independent of its enzymatic activity.
Cell death & disease
Luo W,Li J,Li Z,Lin T,Zhang L,Yang W,Mai Y,Liu R,Chen M,Dai C,Yang H,Lu J,Li H,Guan G,Huang M,Liu P,Li Z
Mon Jul 26 00:00:00 EDT 2021
Pathogenesis of acephalic spermatozoa syndrome caused by SUN5 variant.
Molecular human reproduction
Zhang D,Huang WJ,Chen GY,Dong LH,Tang Y,Zhang H,Li QQ,Mei XY,Wang ZH,Lan FH
Sat May 08 00:00:00 EDT 2021
Ganglioside GD3 regulates dendritic growth in newborn neurons in adult mouse hippocampus via modulation of mitochondrial dynamics.
Journal of neurochemistry
Tang FL,Wang J,Itokazu Y,Yu RK
Mon Mar 01 00:00:00 EST 2021
Urea as a By-Product of Ammonia Metabolism Can Be a Potential Serum Biomarker of Hepatocellular Carcinoma.
Frontiers in cell and developmental biology
Bai C,Wang H,Dong D,Li T,Yu Z,Guo J,Zhou W,Li D,Yan R,Wang L,Wang Z,Li Y,Ren L
Wed Apr 21 00:00:00 EDT 2021
Targeting Mutated p53 Dependency in Triple-Negative Breast Cancer Cells Through CDK7 Inhibition.
Frontiers in oncology
Peng J,Yang M,Bi R,Wang Y,Wang C,Wei X,Zhang Z,Xie X,Wei W
Sat Apr 23 00:00:00 EDT 2022
Stem Cell Factor SOX2 Confers Ferroptosis Resistance in Lung Cancer via Upregulation of SLC7A11.
Cancer research
Wang X,Chen Y,Wang X,Tian H,Wang Y,Jin J,Shan Z,Liu Y,Cai Z,Tong X,Luan Y,Tan X,Luan B,Ge X,Ji H,Jiang X,Wang P
Fri Oct 15 00:00:00 EDT 2021
HN1 promotes tumor growth and metastasis of anaplastic thyroid carcinoma by interacting with STMN1.
Cancer letters
Pan Z,Fang Q,Li L,Zhang Y,Xu T,Liu Y,Zheng X,Tan Z,Huang P,Ge M
Wed Mar 31 00:00:00 EDT 2021
TMEM98, a novel secretory protein, promotes endothelial cell adhesion as well as vascular smooth muscle cell proliferation and migration.
Canadian journal of physiology and pharmacology
Li M,Zhu H,Hu X,Gao F,Hu X,Cui Y,Wei X,Xie C,Lv G,Zhao Y,Gao Y
Sat May 01 00:00:00 EDT 2021
miR-378d suppresses malignant phenotype of ESCC cells through AKT signaling.
Cancer cell international
Peng J,Shi S,Yu J,Liu J,Wei H,Song H,Wang S,Li Z,He S,Li L,Zhang H,Yan Z,Zhao R,Liu Y,Liu Y,Li J,Zhang R,Wang W
Wed Dec 22 00:00:00 EST 2021
Prototheca spp. induce an inflammatory response via mtROS-mediated activation of NF-κB and NLRP3 inflammasome pathways in bovine mammary epithelial cell cultures.
Veterinary research
Zhao W,He F,Barkema HW,Xu S,Gao J,Liu G,Deng Z,Shahid M,Shi Y,Kastelic JP,Han B
Sat Dec 11 00:00:00 EST 2021
Mitochondrial creatine kinase 1 in non-small cell lung cancer progression and hypoxia adaptation.
Respiratory research
Li M,Liu H,Li J,Guo S,Lv Y
Thu Jul 01 00:00:00 EDT 2021
Anticancer Action of Xiaoxianxiong Tang in Non-Small Cell Lung Cancer by Pharmacological Analysis and Experimental Validation.
Evidence-based complementary and alternative medicine : eCAM
Ding R,Jiao L,Sang S,Yin Y,Wang Y,Gong Y,Xu L,Bi L
Fri Dec 24 00:00:00 EST 2021
Aging-associated lncRNAs are evolutionarily conserved and participate in NFκB signaling.
Nature aging
Cai D,Han JJ
Sat May 01 00:00:00 EDT 2021
Screening of a Small Molecule Compound Library Identifies Toosendanin as an Inhibitor Against Bunyavirus and SARS-CoV-2.
Frontiers in pharmacology
Li S,Ye M,Chen Y,Zhang Y,Li J,Liu W,Li H,Peng K
Sat Dec 04 00:00:00 EST 2021
Different roles of BAG3 in cardiac physiological hypertrophy and pathological remodeling.
Translational research : the journal of laboratory and clinical medicine
Jia P,Wu N,Yang H,Guo Y,Guo X,Sun Y
Thu Jul 01 00:00:00 EDT 2021
Effect of Xanthium Strumarium on HIV-1 5'-LTR Transcriptional Activity and Viral Reactivation in Latently Infected Cells.
Frontiers in pharmacology
Chen CJ,Chiu ML,Hung CH,Liang WM,Ho MW,Lin TH,Liu X,Tsang H,Liao CC,Huang SM,Wu YF,Wu YC,Li TM,Tsai FJ,Lin YJ
Tue Aug 24 00:00:00 EDT 2021
A novel CCDC39 mutation causes multiple morphological abnormalities of the flagella in a primary ciliary dyskinesia patient.
Reproductive biomedicine online
Chen D,Liang Y,Li J,Zhang X,Zheng R,Wang X,Zhang H,Shen Y
Mon Nov 01 00:00:00 EDT 2021
Vitellogenin 2 promotes muscle development and stimulates the browning of white fat.
Aging
Li Y,Sun X,Bai Y,Ji Y,Ren H,Yu X,Yan Y,He X,Dong Y,Zhang L,Luo X,Wang H
Tue Oct 05 00:00:00 EDT 2021
Folic Acid Protects Melanocytes from Oxidative Stress via Activation of Nrf2 and Inhibition of HMGB1.
Oxidative medicine and cellular longevity
Du P,Zhang S,Li S,Yang Y,Kang P,Chen J,Gao T,Li C,Zhang Q,Zhang W
Mon Feb 28 00:00:00 EST 2022
S-Propargyl-Cysteine Attenuates Diabetic Cardiomyopathy in db/db Mice Through Activation of Cardiac Insulin Receptor Signaling.
Frontiers in cardiovascular medicine
Li Y,Xie KF,Chang YH,Wang C,Chen Y,Wang MJ,Zhu YC
Tue Oct 05 00:00:00 EDT 2021
Hepatic HuR protects against the pathogenesis of non-alcoholic fatty liver disease by targeting PTEN.
Cell death & disease
Tian M,Wang J,Liu S,Li X,Li J,Yang J,Zhang C,Zhang W
Thu Mar 04 00:00:00 EST 2021
Chlorpromazine, an antipsychotic agent, induces G2/M phase arrest and apoptosis via regulation of the PI3K/AKT/mTOR-mediated autophagy pathways in human oral cancer.
Biochemical pharmacology
Jhou AJ,Chang HC,Hung CC,Lin HC,Lee YC,Liu WT,Han KF,Lai YW,Lin MY,Lee CH
Mon Feb 01 00:00:00 EST 2021
W2476 represses TXNIP transcription via dephosphorylation of FOXO1 at Ser319.
Chemical biology & drug design
Zhong L,Liu Q,Liu Q,Zhang S,Cao Y,Yang D,Wang MW
Sat May 01 00:00:00 EDT 2021
The Phytochemical Scoulerine Inhibits Aurora Kinase Activity to Induce Mitotic and Cytokinetic Defects.
Journal of natural products
Li J,Yan Z,Li H,Shi Q,Ahire V,Zhang S,Nimishetti N,Yang D,Allen TD,Zhang J
Fri Aug 27 00:00:00 EDT 2021
Perfluorooctane sulfonate induces autophagy-dependent lysosomal membrane permeabilization by weakened interaction between tyrosinated alpha-tubulin and spinster 1.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Dong Z,Qiu T,Zhang J,Sha S,Han X,Kang J,Shi X,Sun X,Jiang L,Yang G,Yao X,Ma Y
Mon Nov 01 00:00:00 EDT 2021
Exploring the Prognostic Value, Immune Implication and Biological Function of H2AFY Gene in Hepatocellular Carcinoma.
Frontiers in immunology
Huang Y,Huang S,Ma L,Wang Y,Wang X,Xiao L,Qin W,Li L,Yuan X
Mon Feb 07 00:00:00 EST 2022
Vascular endothelial growth factor alleviates mitochondrial dysfunction and suppression of mitochondrial biogenesis in models of Alzheimer's disease.
The International journal of neuroscience
Liu X,Chu B,Jin S,Li M,Xu Y,Yang H,Feng Z,Bi J,Wang P
Mon Feb 01 00:00:00 EST 2021
Loss of the mitochondrial phosphate carrier SLC25A3 induces remodeling of the cardiac mitochondrial protein acylome.
American journal of physiology. Cell physiology
Peoples JN,Ghazal N,Duong DM,Hardin KR,Manning JR,Seyfried NT,Faundez V,Kwong JQ
Wed Sep 01 00:00:00 EDT 2021
Lower levels of tubulin alpha 1b in the frontal pole in schizophrenia supports a role for changed cytoskeletal dynamics in the aetiology of the disorder.
Psychiatry research
Snelleksz M,Dean B
Wed Sep 01 00:00:00 EDT 2021
Deficiency of Mitochondrial Glycerol 3-Phosphate Dehydrogenase Exacerbates Podocyte Injury and the Progression of Diabetic Kidney Disease.
Diabetes
Qu H,Gong X,Liu X,Zhang R,Wang Y,Huang B,Zhang L,Zheng H,Zheng Y
Tue Jun 01 00:00:00 EDT 2021
Effects and mechanisms of probucol on aging-related hippocampus-dependent cognitive impairment.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Xie Y,Song A,Zhu Y,Jiang A,Peng W,Zhang C,Meng X
Wed Dec 01 00:00:00 EST 2021
MAD2B-mediated cell cycle reentry of podocytes is involved in the pathogenesis of FSGS.
International journal of biological sciences
Bao D,Su H,Lei CT,Tang H,Ye C,Xiong W,He FF,Lin JH,Hammes HP,Zhang C
Mon Dec 20 00:00:00 EST 2021
Drp1 SUMO/deSUMOylation by Senp5 isoforms influences ER tubulation and mitochondrial dynamics to regulate brain development.
iScience
Yamada S,Sato A,Ishihara N,Akiyama H,Sakakibara SI
Fri Dec 17 00:00:00 EST 2021
Drp1 SUMO/deSUMOylation by Senp5 isoforms influences ER tubulation and mitochondrial dynamics to regulate brain development.
iScience
Yamada S,Sato A,Ishihara N,Akiyama H,Sakakibara SI
Fri Dec 17 00:00:00 EST 2021
Drp1 SUMO/deSUMOylation by Senp5 isoforms influences ER tubulation and mitochondrial dynamics to regulate brain development.
iScience
Yamada S,Sato A,Ishihara N,Akiyama H,Sakakibara SI
Fri Dec 17 00:00:00 EST 2021
Drp1 SUMO/deSUMOylation by Senp5 isoforms influences ER tubulation and mitochondrial dynamics to regulate brain development.
iScience
Yamada S,Sato A,Ishihara N,Akiyama H,Sakakibara SI
Fri Dec 17 00:00:00 EST 2021
Glycosylation of Siglec15 promotes immunoescape and tumor growth.
American journal of cancer research
Wang YL,Wei MB,Zhao WW,Feng LL,Yin XK,Bai SM,Wan XB,Hung MC,Zou AZ,Wang MH,Zheng J,Qin C,Fan XJ
Tue Jun 08 00:00:00 EDT 2021
Haploinsufficiency of the TDP43 ubiquitin E3 ligase RNF220 leads to ALS-like motor neuron defects in the mouse.
Journal of molecular cell biology
Ma P,Li Y,Wang H,Mao B
Wed Aug 18 00:00:00 EDT 2021
Inhibitory effect of HTLV-1 infection on the production of B-cell activating factors in established follicular dendritic cell-like cells.
Immunity, inflammation and disease
Takatani A,Nakamura H,Furukawa K,Endo Y,Umeda M,Shimizu T,Nishihata SY,Kitaoka K,Nakamura T,Kawakami A
Wed Sep 01 00:00:00 EDT 2021
MicroRNA-222 Transferred From Semen Extracellular Vesicles Inhibits Sperm Apoptosis by Targeting BCL2L11.
Frontiers in cell and developmental biology
Ding Y,Ding N,Zhang Y,Xie S,Huang M,Ding X,Dong W,Zhang Q,Jiang L
Fri Nov 26 00:00:00 EST 2021
Identification of a Novel Epigenetic Signature CHFR as a Potential Prognostic Gene Involved in Metastatic Clear Cell Renal Cell Carcinoma.
Frontiers in genetics
Chen X,Lin J,Chen Q,Liao X,Wang T,Li S,Mao L,Li Z
Tue Sep 21 00:00:00 EDT 2021
Role of epigenetic m6 A RNA methylation in vascular development: mettl3 regulates vascular development through PHLPP2/mTOR-AKT signaling.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Parial R,Li H,Li J,Archacki S,Yang Z,Wang IZ,Chen Q,Xu C,Wang QK
Sat May 01 00:00:00 EDT 2021
Heme supplementation ameliorates lupus nephritis through rectifying the disorder of splenocytes and alleviating renal inflammation and oxidative damage.
International immunopharmacology
Wu B,Wu Y,Fan C,Feng C,Wang H,Bai F,Zuo J,Tang W
Sat May 01 00:00:00 EDT 2021
Loss of TRIM21 alleviates cardiotoxicity by suppressing ferroptosis induced by the chemotherapeutic agent doxorubicin.
EBioMedicine
Hou K,Shen J,Yan J,Zhai C,Zhang J,Pan JA,Zhang Y,Jiang Y,Wang Y,Lin RZ,Cong H,Gao S,Zong WX
Thu Jul 01 00:00:00 EDT 2021
Selenite Inhibits Notch Signaling in Cells and Mice.
International journal of molecular sciences
Powers M,Liu L,Deemer D,Chen S,Scholl A,Yoshinaga M,Liu Z
Wed Mar 03 00:00:00 EST 2021
Integrated analysis of microRNA and mRNA expression profiling identifies BAIAP3 as a novel target of dysregulated hsa-miR-1972 in age-related white matter lesions.
Aging
Huang WQ,Lin Q,Chen S,Sun L,Chen Q,Yi K,Li Z,Ma Q,Tzeng CM
Tue Feb 09 00:00:00 EST 2021
Immunoprecipitation Strategies to Isolate RIPK1/RIPK3 Complexes in Mouse Macrophages.
Current protocols
Siokas I,Zhang D,Poltorak A,Muendlein H,Degterev A
Tue Jun 01 00:00:00 EDT 2021
Dexmedetomidine post-conditioning attenuates cerebral ischemia following asphyxia cardiac arrest through down-regulation of apoptosis and neuroinflammation in rats.
BMC anesthesiology
Li G,Gu P,Fan D
Mon Jun 28 00:00:00 EDT 2021
Deubiquitinase USP35 restrains STING-mediated interferon signaling in ovarian cancer.
Cell death and differentiation
Zhang J,Chen Y,Chen X,Zhang W,Zhao L,Weng L,Tian H,Wu Z,Tan X,Ge X,Wang P,Fang L
Fri Jan 01 00:00:00 EST 2021
H(+)/Cl(‑) exchange transporter 7 promotes lysosomal acidification‑mediated autophagy in mouse cardiomyocytes.
Molecular medicine reports
Lin J,Wei J,Lv Y,Zhang X,Yi RF,Dai C,Zhang Q,Jia J,Zhang D,Huang Y
Mon Mar 01 00:00:00 EST 2021
Oxidized phospholipids on alkyl-amide scaffold demonstrate anti-endotoxin and endothelial barrier-protective properties.
Free radical biology & medicine
Oskolkova OV,Hodzic A,Karki P,Gesslbauer B,Ke Y,Hofer DC,Bogner-Strauss JG,Galano JM,Oger C,Birukova A,Durand T,Birukov K,Bochkov V
Fri Oct 01 00:00:00 EDT 2021
Tanshinone IIA prevents LPS-induced inflammatory responses in mice via inactivation of succinate dehydrogenase in macrophages.
Acta pharmacologica Sinica
Liu QY,Zhuang Y,Song XR,Niu Q,Sun QS,Li XN,Li N,Liu BL,Huang F,Qiu ZX
Tue Jun 01 00:00:00 EDT 2021
High glucose induces Drp1-mediated mitochondrial fission via the Orai1 calcium channel to participate in diabetic cardiomyocyte hypertrophy.
Cell death & disease
Wu QR,Zheng DL,Liu PM,Yang H,Li LA,Kuang SJ,Lai YY,Rao F,Xue YM,Lin JJ,Liu SX,Chen CB,Deng CY
Fri Feb 26 00:00:00 EST 2021
Expression of Cntn1 is regulated by stress and associated with anxiety and depression phenotypes.
Brain, behavior, and immunity
Li S,Cao W,Zhou S,Ma M,Zhang W,Li F,Li C
Thu Jul 01 00:00:00 EDT 2021
Deltex3 inhibits Epithelial Mesenchymal Transition in Papillary Thyroid Carcinoma via promoting ubiquitination of XRCC5 to regulate the AKT signal pathway.
Journal of Cancer
Wang L,Huang Y,Liu C,Guo M,Ma Z,He J,Wang A,Sun X,Liu Z
Mon Jan 11 00:00:00 EST 2021
Structure-Guided Design of a Small-Molecule Activator of Sirtuin-3 that Modulates Autophagy in Triple Negative Breast Cancer.
Journal of medicinal chemistry
Zhang J,Zou L,Shi D,Liu J,Zhang J,Zhao R,Wang G,Zhang L,Ouyang L,Liu B
Thu Oct 14 00:00:00 EDT 2021
Endometrial cell-derived small extracellular vesicle miR-100-5p promotes functions of trophoblast during embryo implantation.
Molecular therapy. Nucleic acids
Tan Q,Shi S,Liang J,Cao D,Wang S,Wang Z
Fri Mar 05 00:00:00 EST 2021
Folic Acid-Appended Hydroxypropyl-β-Cyclodextrin Exhibits Potent Antitumor Activity in Chronic Myeloid Leukemia Cells via Autophagic Cell Death.
Cancers
Hoshiko T,Kubota Y,Onodera R,Higashi T,Yokoo M,Motoyama K,Kimura S
Thu Oct 28 00:00:00 EDT 2021
Loss of TARBP2 Drives the Progression of Hepatocellular Carcinoma via miR-145-SERPINE1 Axis.
Frontiers in oncology
Li LM,Chen C,Ran RX,Huang JT,Sun HL,Zeng C,Zhang Z,Zhang W,Liu SM
Fri Sep 24 00:00:00 EDT 2021
Reconsolidation of a post-ingestive nutrient memory requires mTOR in the central amygdala.
Molecular psychiatry
Yan Y,Zhang L,Zhu T,Deng S,Ma B,Lv H,Shan X,Cheng H,Jiang K,Zhang T,Meng B,Mei B,Li WG,Li F
Thu Jul 01 00:00:00 EDT 2021
Nicotine-derived NNK induces the stemness enrichment of CRC cells through regulating the balance of DUSP4-ERK1/2 feedback loop.
Ecotoxicology and environmental safety
Chen Y,Wang Q,Cao L,Tang Y,Yao M,Bi H,Huang Y,Sun G,Song J
Sat May 01 00:00:00 EDT 2021
Adenosine A3 receptor activated in H2O2 oxidative stress of primary open-angle glaucoma.
Annals of translational medicine
Zhou Z,Gao Z,Yan W,Zhang Y,Huang J,Xiong K
Thu Apr 01 00:00:00 EDT 2021
A Novel Hydrogen Sulfide Donor Reduces Pilocarpine-Induced Status Epilepticus and Regulates Microglial Inflammatory Profile.
Frontiers in cellular neuroscience
Liu Z,Zhu Z,He Y,Kang Q,Li F,Zhang W,He Y,Lin Y,Huang B,Mo M,Xu P,Zhu X
Tue Dec 21 00:00:00 EST 2021
Single-cell transcriptome atlas of human mesenchymal stem cells exploring cellular heterogeneity.
Clinical and translational medicine
Wang Z,Chai C,Wang R,Feng Y,Huang L,Zhang Y,Xiao X,Yang S,Zhang Y,Zhang X
Wed Dec 01 00:00:00 EST 2021
Sesquiterpene Lactones from Sigesbeckia glabrescens Possessing Potent Anti-inflammatory Activity by Directly Binding to IKKα/β.
Journal of natural products
Gao X,Shen X,Zheng Y,Yang L,Zhang X,Hu G,Jia J,Wang A
Fri Nov 26 00:00:00 EST 2021
CDC20 promotes bone formation via APC/C dependent ubiquitination and degradation of p65.
EMBO reports
Du Y,Zhang M,Liu X,Li Z,Hu M,Tian Y,Lv L,Zhang X,Liu Y,Zhang P,Zhou Y
Mon Sep 06 00:00:00 EDT 2021
HSP90-dependent PUS7 overexpression facilitates the metastasis of colorectal cancer cells by regulating LASP1 abundance.
Journal of experimental & clinical cancer research : CR
Song D,Guo M,Xu S,Song X,Bai B,Li Z,Chen J,An Y,Nie Y,Wu K,Wang S,Zhao Q
Fri May 14 00:00:00 EDT 2021
USP1 Promotes GC Metastasis via Stabilizing ID2.
Disease markers
Li N,Wu L,Zuo X,Luo H,Sheng Y,Yan J
Wed Mar 09 00:00:00 EST 2022
Aberrant expression of PROS1 correlates with human papillary thyroid cancer progression.
PeerJ
Wang J,Lei M,Xu Z
Mon Apr 25 00:00:00 EDT 2022
Changes in the expression of endothelial monocyte‑activating polypeptide II in the rat hippocampus following status epilepticus.
International journal of molecular medicine
Li C,Ma W,Zhao Y,Wang H
Mon Feb 01 00:00:00 EST 2021
USP34 regulates tooth root morphogenesis by stabilizing NFIC.
International journal of oral science
Jiang S,Sheng R,Qi X,Wang J,Guo Y,Yuan Q
Tue Mar 09 00:00:00 EST 2021
33-kDa ANXA3 isoform contributes to hepatocarcinogenesis via modulating ERK, PI3K/Akt-HIF and intrinsic apoptosis pathways.
Journal of advanced research
Guo C,Li N,Dong C,Wang L,Li Z,Liu Q,Ma Q,Greenaway FT,Tian Y,Hao L,Liu S,Sun MZ
Sat May 01 00:00:00 EDT 2021
Ubash3b promotes TPA-mediated suppression of leukemogenesis through accelerated downregulation of PKCδ protein.
Biochimie
Yao Y,Liu W,Gajendran B,Wang C,Zacksenhaus E,Sample KM,Varier KM,Hao X,Ben-David Y
Sat May 01 00:00:00 EDT 2021
TNPO1-Mediated Nuclear Import of FUBP1 Contributes to Tumor Immune Evasion by Increasing NRP1 Expression in Cervical Cancer.
Journal of immunology research
Yang B,Chen J,Teng Y
Mon Nov 29 00:00:00 EST 2021
Copper depletion modulates mitochondrial oxidative phosphorylation to impair triple negative breast cancer metastasis.
Nature communications
Ramchandani D,Berisa M,Tavarez DA,Li Z,Miele M,Bai Y,Lee SB,Ban Y,Dephoure N,Hendrickson RC,Cloonan SM,Gao D,Cross JR,Vahdat LT,Mittal V
Wed Dec 15 00:00:00 EST 2021
The deubiquitinating enzyme ATXN3 promotes the progression of anaplastic thyroid carcinoma by stabilizing EIF5A2.
Molecular and cellular endocrinology
Zhuang S,Xie J,Zhen J,Guo L,Hong Z,Li F,Xu D
Mon Nov 01 00:00:00 EDT 2021
Programmed cell death 5 improves skeletal muscle insulin resistance by inhibiting IRS-1 ubiquitination through stabilization of MDM2.
Life sciences
Li B,Ye J,Liu R,Weng L,Cao Y,Jia S,Xu C,Liu Y,Yan S,Zheng M
Mon Nov 15 00:00:00 EST 2021
Characteristics of inflammation process in monocrotaline-induced pulmonary arterial hypertension in rats.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Tang C,Luo Y,Li S,Huang B,Xu S,Li L
Fri Jan 01 00:00:00 EST 2021
Network-Based Expression Analyses and Experimental Verifications Reveal the Involvement of STUB1 in Acute Kidney Injury.
Frontiers in molecular biosciences
Shi Y,Chen G,Teng J
Fri Jul 16 00:00:00 EDT 2021
Brusatol Inhibits Tumor Growth and Increases the Efficacy of Cabergoline against Pituitary Adenomas.
Oxidative medicine and cellular longevity
Wu Z,Xu Y,Xu J,Lu J,Cai L,Li Q,Wang C,Su Z
Tue Dec 28 00:00:00 EST 2021
Glycoursodeoxycholic acid ameliorates diet-induced metabolic disorders with inhibiting endoplasmic reticulum stress.
Clinical science (London, England : 1979)
Cheng L,Chen T,Guo M,Liu P,Qiao X,Wei Y,She J,Li B,Xi W,Zhou J,Yuan Z,Wu Y,Liu J
Fri Jul 30 00:00:00 EDT 2021
Butyrate alleviates PTZ-induced mitochondrial dysfunction, oxidative stress and neuron apoptosis in mice via Keap1/Nrf2/HO-1 pathway.
Brain research bulletin
Li D,Bai X,Jiang Y,Cheng Y
Mon Mar 01 00:00:00 EST 2021
The inhibitory effect of the deubiquitinase cylindromatosis (CYLD) on inflammatory responses in human gingival fibroblasts.
Oral diseases
Fu YW,Li L,Wang XQ,Zhou Y,Zhu LF,Mei YM,Xu Y
Wed Sep 01 00:00:00 EDT 2021
Bortezomib potentiates antitumor activity of mitoxantrone through dampening Wnt/β-catenin signal pathway in prostate cancer cells.
BMC cancer
Zhang Y,Liu Q,Wei W,Zhang G,Yan S,Dai R,Sun Y,Su D,Lv S,Xia Y,Li J,Li C
Wed Oct 13 00:00:00 EDT 2021
Demethylzeylasteral inhibits proliferation, migration, and invasion through FBXW7/c-Myc axis in gastric cancer.
MedComm
Li Y,Su Y,Zhao Y,Hu X,Zhao G,He J,Wan S,Lü M,Cui H
Wed Sep 01 00:00:00 EDT 2021
A circuit of COCH neurons encodes social-stress-induced anxiety via MTF1 activation of Cacna1h.
Cell reports
Jing W,Zhang T,Liu J,Huang X,Yu Q,Yu H,Zhang Q,Li H,Deng M,Zhu LQ,Du H,Lu Y
Tue Dec 28 00:00:00 EST 2021
Semaphorin 3A promotes the osteogenic differentiation of rat bone marrow-derived mesenchymal stem cells in inflammatory environments by suppressing the Wnt/β-catenin signaling pathway.
Journal of molecular histology
Sun Z,Yan K,Liu S,Yu X,Xu J,Liu J,Li S
Wed Dec 01 00:00:00 EST 2021
The inhibition of colorectal cancer growth by the natural product macrocarpal I.
Free radical biology & medicine
Qi L,Zhang Y,Zhang W,Wang Y,Han Y,Ding Y
Fri Jan 01 00:00:00 EST 2021
GP73 is a TBC-domain Rab GTPase-activating protein contributing to the pathogenesis of non-alcoholic fatty liver disease without obesity.
Nature communications
Peng Y,Zeng Q,Wan L,Ma E,Li H,Yang X,Zhang Y,Huang L,Lin H,Feng J,Xu Y,Li J,Liu M,Liu J,Lin C,Sun Z,Cheng G,Zhang X,Liu J,Li D,Wei M,Mo Y,Mu X,Deng X,Zhang D,Dong S,Huang H,Fang Y,Gao Q,Yang X,Wu F,Zhong H,Wei C
Wed Dec 01 00:00:00 EST 2021
Cilia locally synthesize proteins to sustain their ultrastructure and functions.
Nature communications
Hao K,Chen Y,Yan X,Zhu X
Tue Nov 30 00:00:00 EST 2021
Activation of the STAT3 Signaling Pathway by the RNA-Dependent RNA Polymerase Protein of Arenavirus.
Viruses
Wang Q,Xin Q,Shang W,Wan W,Xiao G,Zhang LK
Tue May 25 00:00:00 EDT 2021
CHD8 safeguards early neuroectoderm differentiation in human ESCs and protects from apoptosis during neurogenesis.
Cell death & disease
Ding S,Lan X,Meng Y,Yan C,Li M,Li X,Chen J,Jiang W
Fri Oct 22 00:00:00 EDT 2021
Central role of TRAP1 in the ameliorative effect of oleanolic acid on the mitochondrial-mediated and endoplasmic reticulum stress-excitated apoptosis induced by ochratoxin A.
Toxicology
Zhang Q,Chen W,Zhang B,Li C,Zhang X,Wang Q,Wang Y,Zhou Q,Li X,Shen XL
Sun Feb 28 00:00:00 EST 2021
Assembly of Cytoplasmic Stress Granules in Placentas in Women with Preeclampsia.
Reproductive sciences (Thousand Oaks, Calif.)
Ma C,Li C,Shao S,Li C,Yu S,Zhao L,Li Y,Zhao S
Fri Oct 01 00:00:00 EDT 2021
Macrophage-Mediated Bone Formation in Scaffolds Modified With MSC-Derived Extracellular Matrix Is Dependent on the Migration Inhibitory Factor Signaling Pathway.
Frontiers in cell and developmental biology
Deng M,Tan J,Dai Q,Luo F,Xu J
Sat Oct 09 00:00:00 EDT 2021
Activation of STING Pathway Contributed to Cisplatin-Induced Cardiac Dysfunction via Promoting the Activation of TNF-α-AP-1 Signal Pathway.
Frontiers in pharmacology
Wang L,Zhang S,Han J,Nie X,Qi Y,Han Y,Chen X,He C
Tue Sep 07 00:00:00 EDT 2021
The RNA binding protein QKI5 suppresses ovarian cancer via downregulating transcriptional coactivator TAZ.
Molecular therapy. Nucleic acids
Liu T,Yang Y,Xie Z,Luo Q,Yang D,Liu X,Zhao H,Wei Q,Liu Y,Li L,Wang Y,Wang F,Yu J,Xu J,Yu J,Yi P
Fri Dec 03 00:00:00 EST 2021
RBM14 Modulates Tubulin Acetylation and Regulates Spindle Morphology During Meiotic Maturation in Mouse Oocytes.
Frontiers in cell and developmental biology
Qin H,Qu Y,Yuan YF,Li YY,Qiao J
Sat Feb 20 00:00:00 EST 2021
RBM14 Modulates Tubulin Acetylation and Regulates Spindle Morphology During Meiotic Maturation in Mouse Oocytes.
Frontiers in cell and developmental biology
Qin H,Qu Y,Yuan YF,Li YY,Qiao J
Sat Feb 20 00:00:00 EST 2021
SARS-CoV-2 spike protein dictates syncytium-mediated lymphocyte elimination.
Cell death and differentiation
Zhang Z,Zheng Y,Niu Z,Zhang B,Wang C,Yao X,Peng H,Franca DN,Wang Y,Zhu Y,Su Y,Tang M,Jiang X,Ren H,He M,Wang Y,Gao L,Zhao P,Shi H,Chen Z,Wang X,Piacentini M,Bian X,Melino G,Liu L,Huang H,Sun Q
Wed Sep 01 00:00:00 EDT 2021
Ggps1 deficiency in the uterus results in dystocia by disrupting uterine contraction.
Journal of molecular cell biology
Sang YJ,Wang Q,Zheng F,Hua Y,Wang XY,Zhang JZ,Li K,Wang HQ,Zhao Y,Zhu MS,Sun HX,Li CJ
Fri May 07 00:00:00 EDT 2021
SARS-CoV-2 inhibits induction of the MHC class I pathway by targeting the STAT1-IRF1-NLRC5 axis.
Nature communications
Yoo JS,Sasaki M,Cho SX,Kasuga Y,Zhu B,Ouda R,Orba Y,de Figueiredo P,Sawa H,Kobayashi KS
Mon Nov 15 00:00:00 EST 2021
Targeting ROCK1/2 blocks cell division and induces mitotic catastrophe in hepatocellular carcinoma.
Biochemical pharmacology
Wu H,Chen Y,Li B,Li C,Guo J,You J,Hu X,Kuang D,Qi S,Liu P,Li L,Xu C
Mon Feb 01 00:00:00 EST 2021
IL20RA signaling enhances stemness and promotes the formation of an immunosuppressive microenvironment in breast cancer.
Theranostics
Gao W,Wen H,Liang L,Dong X,Du R,Zhou W,Zhang X,Zhang C,Xiang R,Li N
Tue Jul 27 00:00:00 EDT 2021
Pragmatic recruitment of memantine as the capping group for the design of HDAC inhibitors: A preliminary attempt to unravel the enigma of glioblastoma.
European journal of medicinal chemistry
Nepali K,Hsu TI,Hsieh CM,Lo WL,Lai MJ,Hsu KC,Lin TE,Chuang JY,Liou JP
Wed May 05 00:00:00 EDT 2021
Effects of andrographolide on renal tubulointersticial injury and fibrosis. Evidence of its mechanism of action.
Phytomedicine : international journal of phytotherapy and phytopharmacology
Liu W,Liang L,Zhang Q,Li Y,Yan S,Tang T,Ren Y,Mo J,Liu F,Chen X,Lan T
Fri Oct 01 00:00:00 EDT 2021
STAT5 inhibitor attenuates atherosclerosis via inhibition of inflammation: the role of STAT5 in atherosclerosis.
American journal of translational research
Wang X,Ding X,Yan J,Lu Z,Cao H,Ni X,Ying Y
Tue Apr 13 00:00:00 EDT 2021
Sevoflurane prevents pulmonary vascular remodeling and right ventricular dysfunction in pulmonary arterial hypertension in rats.
American journal of translational research
Zhai W,Li Y,Luo Y,Gao W,Liu S,Han J,Geng J
Thu Nov 18 00:00:00 EST 2021
High PHD Finger Protein 19 (PHF19) expression predicts poor prognosis in colorectal cancer: a retrospective study.
PeerJ
Li P,Sun J,Ruan Y,Song L
Sun Apr 24 00:00:00 EDT 2022
Discovery of a Dual Tubulin and Poly(ADP-Ribose) Polymerase-1 Inhibitor by Structure-Based Pharmacophore Modeling, Virtual Screening, Molecular Docking, and Biological Evaluation.
Journal of medicinal chemistry
Zheng L,Ren R,Sun X,Zou Y,Shi Y,Di B,Niu MM
Thu Nov 11 00:00:00 EST 2021
Momordin Ic induces G0/1 phase arrest and apoptosis in colon cancer cells by suppressing SENP1/c-MYC signaling pathway.
Journal of pharmacological sciences
Xianjun F,Xirui X,Jie T,Huiwen M,Shaojun Z,Qiaoyun L,Yunxin L,Xuqun S
Sun Aug 01 00:00:00 EDT 2021
Enteric glial cells exert neuroprotection from hyperglycemia-induced damage via Akt/GSK3β pathway.
Neuroreport
Luo P,He WX,Li C,Chang MJ
Wed Jul 07 00:00:00 EDT 2021
AHR mediates the aflatoxin B1 toxicity associated with hepatocellular carcinoma.
Signal transduction and targeted therapy
Zhu Q,Ma Y,Liang J,Wei Z,Li M,Zhang Y,Liu M,He H,Qu C,Cai J,Wang X,Zeng Y,Jiao Y
Mon Aug 09 00:00:00 EDT 2021
Cardioprotective Role of SIRT5 in Response to Acute Ischemia Through a Novel Liver-Cardiac Crosstalk Mechanism.
Frontiers in cell and developmental biology
Zhou B,Xiao M,Hu H,Pei X,Xue Y,Miao G,Wang J,Li W,Du Y,Zhang P,Wei T
Tue Aug 10 00:00:00 EDT 2021
Secreted Frizzled-Related Protein 5 Protects Against Cardiac Rupture and Improves Cardiac Function Through Inhibiting Mitochondrial Dysfunction.
Frontiers in cardiovascular medicine
Huang X,Yan Y,Zheng W,Ma Y,Wang X,Gong W,Nie S
Wed Sep 29 00:00:00 EDT 2021
mTOR regulates PRMT1 expression and mitochondrial mass through STAT1 phosphorylation in hepatic cell.
Biochimica et biophysica acta. Molecular cell research
Zhang X,Li L,Li Y,Li Z,Zhai W,Sun Q,Yang X,Roth M,Lu S
Sat May 01 00:00:00 EDT 2021
The protective effect of isoflurane pretreatment on liver IRI by suppressing noncanonical pyroptosis of liver macrophages.
International immunopharmacology
Lu J,Wang X,Feng Z,Chen Y,Wen D,Liu Z
Fri Oct 01 00:00:00 EDT 2021
AMD1 upregulates hepatocellular carcinoma cells stemness by FTO mediated mRNA demethylation.
Clinical and translational medicine
Bian X,Shi D,Xing K,Zhou H,Lu L,Yu D,Wu W
Mon Mar 01 00:00:00 EST 2021
Apoptotic caspases suppress Mycobacterium bovis-induced IFN-β production in murine macrophage.
The Journal of infection
Song Y,Dong Y,Liao Y,Liang Z,Yao J,Zhou X
Thu Jul 01 00:00:00 EDT 2021
Screening and identification of endometrial proteins as novel potential biomarkers for repeated implantation failure.
PeerJ
Wang C,Feng Y,Zhou WJ,Cheng ZJ,Jiang MY,Zhou Y,Fei XY
Fri Mar 26 00:00:00 EDT 2021
Irradiation-Induced Activated Microglia Affect Brain Metastatic Colonization of NSCLC Cells via miR-9/CDH1 Axis.
OncoTargets and therapy
Jin Y,Kang Y,Peng X,Yang L,Li Q,Mei Q,Chen X,Hu G,Tang Y,Yuan X
Sat Nov 11 00:00:00 EST 2023
Pterostilbene and its nicotinate derivative ameliorated vascular endothelial senescence and elicited endothelium-dependent relaxations via activation of sirtuin 1.
Canadian journal of physiology and pharmacology
Zhang L,Zheng J,Tie X,Lin T,Yang W,Li Z,Zou Y,Guan G,Liu P,Luo W,Li Z
Wed Sep 01 00:00:00 EDT 2021
Triterpene saponins from Guo-gang-long attenuate collagen-induced arthritis via regulating A20 and inhibiting MAPK pathway.
Journal of ethnopharmacology
Xiong H,Luo M,Ju Y,Zhao Z,Zhang M,Xu R,Ren Y,Yang G,Mei Z
Tue Apr 06 00:00:00 EDT 2021
Esomeprazole overcomes paclitaxel-resistance and enhances anticancer effects of paclitaxel by inducing autophagy in A549/Taxol cells.
Cell biology international
Bai Z,Ding N,Ge J,Wang Y,Wang L,Wu N,Wei Q,Xu S,Liu X,Zhou G
Fri Jan 01 00:00:00 EST 2021
Prpf31 is essential for the survival and differentiation of retinal progenitor cells by modulating alternative splicing.
Nucleic acids research
Li J,Liu F,Lv Y,Sun K,Zhao Y,Reilly J,Zhang Y,Tu J,Yu S,Liu X,Qin Y,Huang Y,Gao P,Jia D,Chen X,Han Y,Shu X,Luo D,Tang Z,Liu M
Fri Feb 26 00:00:00 EST 2021
Knockdown of ASH1L methyltransferase induced apoptosis inhibiting proliferation and H3K36 methylation in bovine cumulus cells.
Theriogenology
Cui LX,Tian YQ,Hao HS,Zou HY,Pang YW,Zhao SJ,Zhao XM,Zhu HB,Du WH
Mon Feb 01 00:00:00 EST 2021
Impaired Retrograde Transport Due to Lack of TBC1D5 Contributes to the Trafficking Defect of Lysosomal Cathepsins in Ischemic/Hypoxic Cardiomyocytes.
Frontiers in cardiovascular medicine
Cui L,Zhang Q,Huang Y,Yang L,Zhang J,Jiang X,Jia J,Lv Y,Zhang D,Huang Y
Tue Jan 11 00:00:00 EST 2022
Impaired Retrograde Transport Due to Lack of TBC1D5 Contributes to the Trafficking Defect of Lysosomal Cathepsins in Ischemic/Hypoxic Cardiomyocytes.
Frontiers in cardiovascular medicine
Cui L,Zhang Q,Huang Y,Yang L,Zhang J,Jiang X,Jia J,Lv Y,Zhang D,Huang Y
Tue Jan 11 00:00:00 EST 2022
Impaired Retrograde Transport Due to Lack of TBC1D5 Contributes to the Trafficking Defect of Lysosomal Cathepsins in Ischemic/Hypoxic Cardiomyocytes.
Frontiers in cardiovascular medicine
Cui L,Zhang Q,Huang Y,Yang L,Zhang J,Jiang X,Jia J,Lv Y,Zhang D,Huang Y
Tue Jan 11 00:00:00 EST 2022
SIRT1 inhibitors mitigate radiation-induced GI syndrome by enhancing intestinal-stem-cell survival.
Cancer letters
Fu G,Chen S,Liang L,Li X,Tang P,Rao X,Pan M,Xu X,Li Y,Yao Y,Zhou Y,Gao J,Mo S,Cai S,Peng J,Zhang Z,Clevers H,Gao J,Hua G
Wed Mar 31 00:00:00 EDT 2021
PKD2 deficiency suppresses amino acid biosynthesis in ADPKD by impairing the PERK-TBL2-eIF2α-ATF4 pathway.
Biochemical and biophysical research communications
Zhou X,Xiong H,Lu Y,Geng M,Lu Y,Zhang L,Zhu X
Mon Jul 05 00:00:00 EDT 2021
Treg deficiency-mediated TH 1 response causes human premature ovarian insufficiency through apoptosis and steroidogenesis dysfunction of granulosa cells.
Clinical and translational medicine
Jiao X,Zhang X,Li N,Zhang D,Zhao S,Dang Y,Zanvit P,Jin W,Chen ZJ,Chen W,Qin Y
Tue Jun 01 00:00:00 EDT 2021
19-Hydroxybufalin inhibits non-small cell lung cancer cell proliferation and promotes cell apoptosis via the Wnt/β-catenin pathway.
Experimental hematology & oncology
Yu W,Zhang X,Zhang W,Xiong M,Lin Y,Chang M,Xu L,Lu Y,Liu Y,Zhang J
Mon Oct 25 00:00:00 EDT 2021
[ANXA8 Regulates Proliferation of Human Non-Small Lung Cancer Cells A549 via EGFR-AKT-mTOR Signaling Pathway].
Molekuliarnaia biologiia
Zhou GZ,Sun YH,Shi YY,Zhang Q,Zhang L,Cui LQ,Sun GC
Fri Oct 22 00:00:00 EDT 2021
UFL1 regulates milk protein and fat synthesis-related gene expression of bovine mammary epithelial cells probably via the mTOR signaling pathway.
In vitro cellular & developmental biology. Animal
Li C,Li L,Ali I,Kuang M,Wang X,Wang G
Sat May 01 00:00:00 EDT 2021
METTL3-Mediated m6 A mRNA Methylation Modulates Tooth Root Formation by Affecting NFIC Translation.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research
Sheng R,Wang Y,Wu Y,Wang J,Zhang S,Li Q,Zhang D,Qi X,Xiao Q,Jiang S,Yuan Q
Mon Feb 01 00:00:00 EST 2021
Human splenic TER cells: A relevant prognostic factor acting via the artemin-GFRα3-ERK pathway in pancreatic ductal adenocarcinoma.
International journal of cancer
Li TJ,Li H,Zhang WH,Xu SS,Jiang W,Li S,Gao HL,Han X,Xu HX,Wu CT,Wang WQ,Yu XJ,Liu L
Thu Apr 01 00:00:00 EDT 2021
Glucagon-like Peptide-1 Improves Fatty Liver and Enhances Thermogenesis in Brown Adipose Tissue via Inhibiting BMP4-Related Signaling Pathway in High-Fat-Diet-Induced Obese Mice.
International journal of endocrinology
Wang X,Ma B,Chen J,You H,Sheng C,Yang P,Qu S
Sat Apr 23 00:00:00 EDT 2022
Inhibition of MAD2B alleviates venous neointimal formation by suppressing VSMCs proliferation and migration.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Song AN,Yin XJ,Gao P,Tang H,Meng XF,Zhang C
Mon Nov 01 00:00:00 EDT 2021
ZC3H15 Correlates with a Poor Prognosis and Tumor Progression in Melanoma.
BioMed research international
Li Q,Hou J,Guo C,Zhang Y,Liu L,Hu H,Shi S,Ji Y,Guo L,Shi Y,Liu Y,Cui H
Fri Jan 14 00:00:00 EST 2022
K29-linked ubiquitin signaling regulates proteotoxic stress response and cell cycle.
Nature chemical biology
Yu Y,Zheng Q,Erramilli SK,Pan M,Park S,Xie Y,Li J,Fei J,Kossiakoff AA,Liu L,Zhao M
Sun Aug 01 00:00:00 EDT 2021
Yeast Two-Hybrid Screen Identifies PKA-Riα Interacting Proteins during Mouse Spermiogenesis.
Genes
Shi K,Yang L,Zhuang X,Zhang L,Qi H
Tue Nov 30 00:00:00 EST 2021
PEX5 prevents cardiomyocyte hypertrophy via suppressing the redox-sensitive signaling pathways MAPKs and STAT3.
European journal of pharmacology
Wang M,Li J,Ding Y,Cai S,Li Z,Liu P
Sun Sep 05 00:00:00 EDT 2021
Cold-Induced RNA-Binding Protein Promotes Glucose Metabolism and Reduces Apoptosis by Increasing AKT Phosphorylation in Mouse Skeletal Muscle Under Acute Cold Exposure.
Frontiers in molecular biosciences
Liu Y,Liu P,Hu Y,Cao Y,Lu J,Yang Y,Lv H,Lian S,Xu B,Li S
Tue Aug 17 00:00:00 EDT 2021
SARS-CoV-2 envelope protein causes acute respiratory distress syndrome (ARDS)-like pathological damages and constitutes an antiviral target.
Cell research
Xia B,Shen X,He Y,Pan X,Liu FL,Wang Y,Yang F,Fang S,Wu Y,Duan Z,Zuo X,Xie Z,Jiang X,Xu L,Chi H,Li S,Meng Q,Zhou H,Zhou Y,Cheng X,Xin X,Jin L,Zhang HL,Yu DD,Li MH,Feng XL,Chen J,Jiang H,Xiao G,Zheng YT,Zhang LK,Shen J,Li J,Gao Z
Sun Aug 01 00:00:00 EDT 2021
SARS-CoV-2 triggers inflammatory responses and cell death through caspase-8 activation.
Signal transduction and targeted therapy
Li S,Zhang Y,Guan Z,Li H,Ye M,Chen X,Shen J,Zhou Y,Shi ZL,Zhou P,Peng K
Fri Oct 09 00:00:00 EDT 2020
Uncoupling DNA damage from chromatin damage to detoxify doxorubicin.
Proceedings of the National Academy of Sciences of the United States of America
Qiao X,van der Zanden SY,Wander DPA,Borràs DM,Song JY,Li X,van Duikeren S,van Gils N,Rutten A,van Herwaarden T,van Tellingen O,Giacomelli E,Bellin M,Orlova V,Tertoolen LGJ,Gerhardt S,Akkermans JJ,Bakker JM,Zuur CL,Pang B,Smits AM,Mummery CL,Smit L,Arens R,Li J,Overkleeft HS,Neefjes J
Tue Jun 30 00:00:00 EDT 2020
Ophiopogonin D promotes bone regeneration by stimulating CD31hi EMCNhi vessel formation.
Cell proliferation
Yang M,Li CJ,Xiao Y,Guo Q,Huang Y,Su T,Luo XH,Jiang TJ
Sun Mar 01 00:00:00 EST 2020
Establishment of cell lines with porcine spermatogonial stem cell properties.
Journal of animal science and biotechnology
Zheng Y,Feng T,Zhang P,Lei P,Li F,Zeng W
Thu Apr 14 00:00:00 EDT 2022
PCDH7 Inhibits the Formation of Homotypic Cell-in-Cell Structure.
Frontiers in cell and developmental biology
Wang C,Chen A,Ruan B,Niu Z,Su Y,Qin H,Zheng Y,Zhang B,Gao L,Chen Z,Huang H,Wang X,Sun Q
Mon Sep 28 00:00:00 EDT 2020
E6‑regulated overproduction of prostaglandin E2 may inhibit migration of dendritic cells in human papillomavirus 16‑positive cervical lesions.
International journal of oncology
Huang J,Diao G,Zhang Q,Chen Y,Han J,Guo J
Wed Apr 01 00:00:00 EDT 2020
Hypermethylation of UCHL1 Promotes Metastasis of Nasopharyngeal Carcinoma by Suppressing Degradation of Cortactin (CTTN).
Cells
Zhao Y,Lei Y,He SW,Li YQ,Wang YQ,Hong XH,Liang YL,Li JY,Chen Y,Luo WJ,Zhang PP,Yang XJ,He QM,Ma J,Liu N,Tang LL
Thu Feb 27 00:00:00 EST 2020
PIM1 inhibitor synergizes the anti-tumor effect of osimertinib via STAT3 dephosphorylation in EGFR-mutant non-small cell lung cancer.
Annals of translational medicine
Sun Z,Zeng L,Zhang M,Zhang Y,Yang N
Sun Mar 01 00:00:00 EST 2020
Role of Clostridium perfringens Enterotoxin on YAP Activation in Colonic Sessile Serrated Adenoma/ Polyps with Dysplasia.
International journal of molecular sciences
Fujiwara-Tani R,Fujii K,Mori S,Kishi S,Sasaki T,Ohmori H,Nakashima C,Kawahara I,Nishiguchi Y,Mori T,Sho M,Kondoh M,Luo Y,Kuniyasu H
Thu May 28 00:00:00 EDT 2020
Design, synthesis and biological evaluation of homoerythrina alkaloid derivatives bearing a triazole moiety as PARP-1 inhibitors and as potential antitumor drugs.
Bioorganic chemistry
Li S,Li XY,Zhang TJ,Kamara MO,Liang JW,Zhu J,Meng FH
Wed Jan 01 00:00:00 EST 2020
ASPH Regulates Osteogenic Differentiation and Cellular Senescence of BMSCs.
Frontiers in cell and developmental biology
Peng H,Guo Q,Xiao Y,Su T,Jiang TJ,Guo LJ,Wang M
Tue Oct 06 00:00:00 EDT 2020
Loss of ZC4H2 and RNF220 Inhibits Neural Stem Cell Proliferation and Promotes Neuronal Differentiation.
Cells
Zhang L,Ye M,Zhu L,Cha J,Li C,Yao YG,Mao B
Wed Jul 01 00:00:00 EDT 2020
Mass spectrometry-based proteomic capture of proteins bound to the MACC1 promoter in colon cancer.
Clinical & experimental metastasis
Huang Y,Xiang Y,Xie Z,Cai Y,Yang Q,Huang H,Chen Z,Xiao Z,He Q
Sat Aug 01 00:00:00 EDT 2020
RGS12 is required for the maintenance of mitochondrial function during skeletal development.
Cell discovery
Yuan G,Yang S,Liu M,Yang S
Wed Nov 17 00:00:00 EST 2021
DNA methylation‑regulated miR‑155‑5p depresses sensitivity of esophageal carcinoma cells to radiation and multiple chemotherapeutic drugs via suppression of MAP3K10.
Oncology reports
Luo W,Zhang H,Liang X,Xia R,Deng H,Yi Q,Lv L,Qian L
Fri May 01 00:00:00 EDT 2020
Dnajb8, a target gene of SOX30, is dispensable for male fertility in mice.
PeerJ
Wang F,Kong S,Hu X,Li X,Xu B,Yue Q,Fu K,Ye L,Bai S
Tue Apr 19 00:00:00 EDT 2022
Autophagy suppression of trophoblast cells induces pregnancy loss by activating decidual NK cytotoxicity and inhibiting trophoblast invasion.
Cell communication and signaling : CCS
Tan HX,Yang SL,Li MQ,Wang HY
Tue May 12 00:00:00 EDT 2020
Sulforaphane-cysteine inhibited migration and invasion via enhancing mitophagosome fusion to lysosome in human glioblastoma cells.
Cell death & disease
Zhou Y,Wang Y,Wu S,Yan Y,Hu Y,Zheng Z,Li J,Wu W
Thu Oct 01 00:00:00 EDT 2020
Sulforaphane-cysteine inhibited migration and invasion via enhancing mitophagosome fusion to lysosome in human glioblastoma cells.
Cell death & disease
Zhou Y,Wang Y,Wu S,Yan Y,Hu Y,Zheng Z,Li J,Wu W
Thu Oct 01 00:00:00 EDT 2020
The identification and characterisation of autophagy inhibitors from the published kinase inhibitor sets.
The Biochemical journal
Zachari M,Rainard JM,Pandarakalam GC,Robinson L,Gillespie J,Rajamanickam M,Hamon V,Morrison A,Ganley IG,McElroy SP
Fri Feb 28 00:00:00 EST 2020
5-(3,4,5-trimethoxybenzoyl)-4-methyl-2-(p-tolyl) imidazol (BZML) targets tubulin and DNA to induce anticancer activity and overcome multidrug resistance in colorectal cancer cells.
Chemico-biological interactions
Bai Z,Liu X,Guan Q,Ding N,Wei Q,Tong B,Zhao M,Zhang W,Ma L
Sun Jan 05 00:00:00 EST 2020
Elevated myocardial SORBS2 and the underlying implications in left ventricular noncompaction cardiomyopathy.
EBioMedicine
Li C,Liu F,Liu S,Pan H,Du H,Huang J,Xie Y,Li Y,Zhao R,Wei Y
Sun Mar 01 00:00:00 EST 2020
Naked antisense double-stranded DNA oligonucleotide efficiently suppresses BCR-ABL positive leukemic cells.
Investigational new drugs
Hoshiko T,Kubota Y,Akisawa T,Watanabe T,Tanigawara K,Yano J,Kimura S
Sat Aug 01 00:00:00 EDT 2020
Platycodon grandifloras polysaccharides inhibit mitophagy injury induced by Cr (VI) in DF-1 cells.
Ecotoxicology and environmental safety
Hao J,Song Y,Tian B,Qi C,Li L,Wang L,Xing Y,Zhao X,Liu J
Thu Oct 01 00:00:00 EDT 2020
Hypoxia-induced MFAP5 Promotes Tumor Migration and Invasion via AKT Pathway in Head and Neck Squamous Cell Carcinoma.
Journal of Cancer
Xu Q,Chang H,Tian X,Lou C,Ma H,Yang X
Mon Sep 28 00:00:00 EDT 2020
Antibiotic tigecycline inhibits cell proliferation, migration and invasion via down-regulating CCNE2 in pancreatic ductal adenocarcinoma.
Journal of cellular and molecular medicine
Yang J,Dong Z,Ren A,Fu G,Zhang K,Li C,Wang X,Cui H
Wed Apr 01 00:00:00 EDT 2020
Epigenetic Inactivation of α-Internexin Accelerates Microtubule Polymerization in Colorectal Cancer.
Cancer research
Li Y,Bai L,Yu H,Cai D,Wang X,Huang B,Peng S,Huang M,Cao G,Kaz AM,Grady WM,Wang J,Luo Y
Tue Dec 01 00:00:00 EST 2020
Epigenetic Inactivation of α-Internexin Accelerates Microtubule Polymerization in Colorectal Cancer.
Cancer research
Li Y,Bai L,Yu H,Cai D,Wang X,Huang B,Peng S,Huang M,Cao G,Kaz AM,Grady WM,Wang J,Luo Y
Tue Dec 01 00:00:00 EST 2020
Transient inhibition of rDNA transcription in donor cells improves ribosome biogenesis and preimplantation development of embryos derived from somatic cell nuclear transfer.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Liao C,Pang N,Liu Z,Lei L
Mon Jun 01 00:00:00 EDT 2020
Overexpression of B7-H4 promotes renal cell carcinoma progression by recruiting tumor-associated neutrophils via upregulation of CXCL8.
Oncology letters
Li A,Zhang N,Zhao Z,Chen Y,Zhang L
Sat Aug 01 00:00:00 EDT 2020
Second-order optimized regularized structured illumination microscopy (sorSIM) for high-quality and rapid super resolution image reconstruction with low signal level.
Optics express
Yu W,Li Y,Jooken S,Deschaume O,Liu F,Wang S,Bartic C
Mon May 25 00:00:00 EDT 2020
Lysosomal dysfunction and autophagy blockade contribute to autophagy-related cancer suppressing peptide-induced cytotoxic death of cervical cancer cells through the AMPK/mTOR pathway.
Journal of experimental & clinical cancer research : CR
Yang Y,Wang Q,Song D,Zen R,Zhang L,Wang Y,Yang H,Zhang D,Jia J,Zhang J,Wang J
Tue Sep 22 00:00:00 EDT 2020
Heme Oxygenase-1 Suppresses Wnt Signaling Pathway in Nonalcoholic Steatohepatitis-Related Liver Fibrosis.
BioMed research international
Du J,Ren W,Zhang Q,Fu N,Han F,Cui P,Li W,Kong L,Zhao S,Wang R,Zhang Y,Yang L,Kong L,Nan Y
Fri Mar 12 00:00:00 EST 2021
DUSP9 Suppresses Proliferation and Migration of Clear Cell Renal Cell Carcinoma via the mTOR Pathway.
OncoTargets and therapy
Luo J,Luo X,Liu X,Fang Z,Xu J,Li L
Wed Apr 13 00:00:00 EDT 2022
Protein acetylation derepresses Serotonin Synthesis to potentiate Pancreatic Beta-Cell Function through HDAC1-PKA-Tph1 signaling.
Theranostics
Zhang Y,Wang S,Zhang L,Zhou F,Zhu K,Zhu Q,Liu Q,Liu Y,Jiang L,Ning G,Bi Y,Zhou L,Wang X
Mon May 03 00:00:00 EDT 2021
Gentiopicroside activates the bile acid receptor Gpbar1 (TGR5) to repress NF-kappaB pathway and ameliorate diabetic nephropathy.
Pharmacological research
Xiao H,Sun X,Liu R,Chen Z,Lin Z,Yang Y,Zhang M,Liu P,Quan S,Huang H
Wed Jan 01 00:00:00 EST 2020
Poricoic acid A as a modulator of TPH-1 expression inhibits renal fibrosis via modulating protein stability of β-catenin and β-catenin-mediated transcription.
Therapeutic advances in chronic disease
Chen DQ,Wu XQ,Chen L,Hu HH,Wang YN,Zhao YY
Sun Apr 17 00:00:00 EDT 2022
Human alkaline ceramidase 2 promotes the growth, invasion, and migration of hepatocellular carcinoma cells via sphingomyelin phosphodiesterase acid-like 3B.
Cancer science
Liu B,Xiao J,Dong M,Qiu Z,Jin J
Wed Jul 01 00:00:00 EDT 2020
PhospholipaseCγ1/calcium-dependent membranous localization of Gsdmd-N drives endothelial pyroptosis, contributing to lipopolysaccharide-induced fatal outcome.
American journal of physiology. Heart and circulatory physiology
Liu H,Tang D,Zhou X,Yang X,Chen AF
Tue Dec 01 00:00:00 EST 2020
ING4 alleviated lipopolysaccharide-induced inflammation by regulating the NF-κB pathway via a direct interaction with SIRT1.
Immunology and cell biology
Yang Y,Liu Y,He X,Yang F,Han S,Qin A,Wu G,Liu M,Li Z,Wang J,Yang X,Hu D
Sat Feb 01 00:00:00 EST 2020
Dichloroacetate restores colorectal cancer chemosensitivity through the p53/miR-149-3p/PDK2-mediated glucose metabolic pathway.
Oncogene
Liang Y,Hou L,Li L,Li L,Zhu L,Wang Y,Huang X,Hou Y,Zhu D,Zou H,Gu Y,Weng X,Wang Y,Li Y,Wu T,Yao M,Gross I,Gaiddon C,Luo M,Wang J,Meng X
Wed Jan 01 00:00:00 EST 2020
MicroRNA-384 Inhibits the Progression of Papillary Thyroid Cancer by Targeting PRKACB.
BioMed research international
Wang Y,Wang B,Zhou H,Zhang X,Qian X,Cui J
Mon Sep 21 00:00:00 EDT 2020
Poricoic acid A activates AMPK to attenuate fibroblast activation and abnormal extracellular matrix remodelling in renal fibrosis.
Phytomedicine : international journal of phytotherapy and phytopharmacology
Chen DQ,Wang YN,Vaziri ND,Chen L,Hu HH,Zhao YY
Wed Jul 01 00:00:00 EDT 2020
Mechanical Ring Interfaces between Adherens Junction and Contractile Actomyosin to Coordinate Entotic Cell-in-Cell Formation.
Cell reports
Wang M,Niu Z,Qin H,Ruan B,Zheng Y,Ning X,Gu S,Gao L,Chen Z,Wang X,Huang H,Ma L,Sun Q
Tue Aug 25 00:00:00 EDT 2020
Mechanical Ring Interfaces between Adherens Junction and Contractile Actomyosin to Coordinate Entotic Cell-in-Cell Formation.
Cell reports
Wang M,Niu Z,Qin H,Ruan B,Zheng Y,Ning X,Gu S,Gao L,Chen Z,Wang X,Huang H,Ma L,Sun Q
Tue Aug 25 00:00:00 EDT 2020
Clinically relevant high levels of human C-reactive protein induces endothelial dysfunction and hypertension by inhibiting the AMPK-eNOS axis.
Clinical science (London, England : 1979)
Cheng L,Wang L,Guo M,He J,Deng Y,Liu J,Wei Y,Wang C,Zhou J,Ma L,Song Q,Yuan Z,Wu Y
Fri Jul 17 00:00:00 EDT 2020
EHF promotes colorectal carcinoma progression by activating TGF-β1 transcription and canonical TGF-β signaling.
Cancer science
Wang L,Ai M,Nie M,Zhao L,Deng G,Hu S,Han Y,Zeng W,Wang Y,Yang M,Wang S
Wed Jul 01 00:00:00 EDT 2020
Ambroxol Improves Neuronal Survival and Reduces White Matter Damage through Suppressing Endoplasmic Reticulum Stress in Microglia after Intracerebral Hemorrhage.
BioMed research international
Jiang X,Zhang J,Kou B,Zhang C,Zhong J,Fang X,Huang X,Zhang X,Xie F,Hu Q,Ge H,Yu A
Mon Jan 11 00:00:00 EST 2021
Consumption of a Blueberry-Enriched Diet by Women for 6 Weeks Alters Determinants of Human Muscle Progenitor Cell Function.
The Journal of nutrition
Blum JE,Gheller BJ,Hwang S,Bender E,Gheller M,Thalacker-Mercer AE
Tue Sep 01 00:00:00 EDT 2020
Modification of PLGA Scaffold by MSC-Derived Extracellular Matrix Combats Macrophage Inflammation to Initiate Bone Regeneration via TGF-β-Induced Protein.
Advanced healthcare materials
Deng M,Tan J,Hu C,Hou T,Peng W,Liu J,Yu B,Dai Q,Zhou J,Yang Y,Dong R,Ruan C,Dong S,Xu J
Wed Jul 01 00:00:00 EDT 2020
Regulation of epidermal differentiation through KDF1-mediated deubiquitination of IKKα.
EMBO reports
Li Y,Tang L,Yue J,Gou X,Lin A,Weatherbee SD,Wu X
Wed May 06 00:00:00 EDT 2020
BRCA1 protects cardiac microvascular endothelial cells against irradiation by regulating p21-mediated cell cycle arrest.
Life sciences
Zeng ZM,Du HY,Xiong L,Zeng XL,Zhang P,Cai J,Huang L,Liu AW
Sun Mar 01 00:00:00 EST 2020
RNF6 promotes myeloma cell proliferation and survival by inducing glucocorticoid receptor polyubiquitination.
Acta pharmacologica Sinica
Ren Y,Xu X,Mao CY,Han KK,Xu YJ,Cao BY,Zhang ZB,Sethi G,Tang XW,Mao XL
Sun Mar 01 00:00:00 EST 2020
Therapeutic efficacy of antisense oligonucleotides in mouse models of CLN3 Batten disease.
Nature medicine
Centa JL,Jodelka FM,Hinrich AJ,Johnson TB,Ochaba J,Jackson M,Duelli DM,Weimer JM,Rigo F,Hastings ML
Tue Sep 01 00:00:00 EDT 2020
Asiaticoside Inhibits Neuronal Apoptosis and Promotes Functional Recovery After Spinal Cord Injury in Rats.
Journal of molecular neuroscience : MN
Fan L,Li X,Liu T
Tue Dec 01 00:00:00 EST 2020
Kaempferol attenuated cisplatin-induced cardiac injury via inhibiting STING/NF-κB-mediated inflammation.
American journal of translational research
Qi Y,Ying Y,Zou J,Fang Q,Yuan X,Cao Y,Cai Y,Fu S
Thu Jan 14 00:00:00 EST 2021
Dependence of PINK1 accumulation on mitochondrial redox system.
Aging cell
Gao F,Zhang Y,Hou X,Tao Z,Ren H,Wang G
Tue Sep 01 00:00:00 EDT 2020
Water treadmill training attenuates blood-spinal cord barrier disruption in rats by promoting angiogenesis and inhibiting matrix metalloproteinase-2/9 expression following spinal cord injury.
Fluids and barriers of the CNS
Ying X,Xie Q,Li S,Yu X,Zhou K,Yue J,Chen X,Tu W,Yang G,Jiang S
Wed Nov 25 00:00:00 EST 2020
MKL1 mediates TGF-β-induced CTGF transcription to promote renal fibrosis.
Journal of cellular physiology
Mao L,Liu L,Zhang T,Wu X,Zhang T,Xu Y
Fri May 01 00:00:00 EDT 2020
High-Intensity Interval Training Restores Glycolipid Metabolism and Mitochondrial Function in Skeletal Muscle of Mice With Type 2 Diabetes.
Frontiers in endocrinology
Zheng L,Rao Z,Guo Y,Chen P,Xiao W
Wed May 19 00:00:00 EDT 2021
Proanthocyanidins Promote Osteogenic Differentiation of Human Periodontal Ligament Fibroblasts in Inflammatory Environment Via Suppressing NF-κB Signal Pathway.
Inflammation
Huang J,Liu L,Jin S,Zhang Y,Zhang L,Li S,Song A,Yang P
Mon Jun 01 00:00:00 EDT 2020
Rnf220/Zc4h2-mediated monoubiquitylation of Phox2 is required for noradrenergic neuron development.
Development (Cambridge, England)
Song NN,Ma P,Zhang Q,Zhang L,Wang H,Zhang L,Zhu L,He CH,Mao B,Ding YQ
Mon Mar 30 00:00:00 EDT 2020
Induction of ASC pyroptosis requires gasdermin D or caspase-1/11-dependent mediators and IFNβ from pyroptotic macrophages.
Cell death & disease
Zhang C,Zhao C,Chen X,Tao R,Wang S,Meng G,Liu X,Shao C,Su X
Thu Jun 18 00:00:00 EDT 2020
Modulation of Increased mGluR1 Signaling by RGS8 Protects Purkinje Cells From Dendritic Reduction and Could Be a Common Mechanism in Diverse Forms of Spinocerebellar Ataxia.
Frontiers in cell and developmental biology
Wu QW,Kapfhammer JP
Wed Feb 10 00:00:00 EST 2021
SYTL4 downregulates microtubule stability and confers paclitaxel resistance in triple-negative breast cancer.
Theranostics
Liu XY,Jiang W,Ma D,Ge LP,Yang YS,Gou ZC,Xu XE,Shao ZM,Jiang YZ
Mon May 31 00:00:00 EDT 2021
SYTL4 downregulates microtubule stability and confers paclitaxel resistance in triple-negative breast cancer.
Theranostics
Liu XY,Jiang W,Ma D,Ge LP,Yang YS,Gou ZC,Xu XE,Shao ZM,Jiang YZ
Mon May 31 00:00:00 EDT 2021
SYTL4 downregulates microtubule stability and confers paclitaxel resistance in triple-negative breast cancer.
Theranostics
Liu XY,Jiang W,Ma D,Ge LP,Yang YS,Gou ZC,Xu XE,Shao ZM,Jiang YZ
Mon May 31 00:00:00 EDT 2021
Ophiocordyceps lanpingensis polysaccharides attenuate pulmonary fibrosis in mice.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Zhou S,Zhou Y,Yu J,Du Y,Tan Y,Ke Y,Wang J,Han B,Ge F
Mon Jun 01 00:00:00 EDT 2020
Wnt5a-induced M2 polarization of tumor-associated macrophages via IL-10 promotes colorectal cancer progression.
Cell communication and signaling : CCS
Liu Q,Yang C,Wang S,Shi D,Wei C,Song J,Lin X,Dou R,Bai J,Xiang Z,Huang S,Liu K,Xiong B
Mon Mar 30 00:00:00 EDT 2020
JARID1B promotes colorectal cancer proliferation and Wnt/β-catenin signaling via decreasing CDX2 level.
Cell communication and signaling : CCS
Huang D,Xiao F,Hao H,Hua F,Luo Z,Huang Z,Li Q,Chen S,Cheng X,Zhang X,Fang W,Hu X,Liu F
Tue Oct 27 00:00:00 EDT 2020
C9orf72 associates with inactive Rag GTPases and regulates mTORC1-mediated autophagosomal and lysosomal biogenesis.
Aging cell
Wang M,Wang H,Tao Z,Xia Q,Hao Z,Prehn JHM,Zhen X,Wang G,Ying Z
Wed Apr 01 00:00:00 EDT 2020
EphA5 Silencing Increases the Radiosensitivity of ESCC Cells Through ATM-Dependent Pathway.
Cancer management and research
Zhang R,Han D,Li L,Luo W,Liu J,Qian L
Sun Apr 17 00:00:00 EDT 2022
Neurotoxicity and biomarkers of zinc oxide nanoparticles in main functional brain regions and dopaminergic neurons.
The Science of the total environment
Liu H,Yang H,Fang Y,Li K,Tian L,Liu X,Zhang W,Tan Y,Lai W,Bian L,Lin B,Xi Z
Tue Feb 25 00:00:00 EST 2020
Modulation of SIRT1-mediated signaling cascades in the liver contributes to the amelioration of nonalcoholic steatohepatitis in high fat fed middle-aged LDL receptor knockout mice by dihydromyricetin.
Biochemical pharmacology
Zeng Y,Hua YQ,Wang W,Zhang H,Xu XL
Fri May 01 00:00:00 EDT 2020
The histone modification reader ZCWPW1 links histone methylation to PRDM9-induced double-strand break repair.
eLife
Huang T,Yuan S,Gao L,Li M,Yu X,Zhan J,Yin Y,Liu C,Zhang C,Lu G,Li W,Liu J,Chen ZJ,Liu H
Wed May 06 00:00:00 EDT 2020
Loss of hepatic aldolase B activates Akt and promotes hepatocellular carcinogenesis by destabilizing the Aldob/Akt/PP2A protein complex.
PLoS biology
He X,Li M,Yu H,Liu G,Wang N,Yin C,Tu Q,Narla G,Tao Y,Cheng S,Yin H
Tue Dec 01 00:00:00 EST 2020
GDF15 knockdown promotes erastin-induced ferroptosis by decreasing SLC7A11 expression.
Biochemical and biophysical research communications
Chen L,Qiao L,Bian Y,Sun X
Thu May 28 00:00:00 EDT 2020
Dietary Supplemented Curcumin Improves Meat Quality and Antioxidant Status of Intrauterine Growth Retardation Growing Pigs via Nrf2 Signal Pathway.
Animals : an open access journal from MDPI
Zhang L,Zhang J,Yan E,He J,Zhong X,Zhang L,Wang C,Wang T
Tue Mar 24 00:00:00 EDT 2020
Inhibition of UCHL1 by LDN-57444 attenuates Ang II-Induced atrial fibrillation in mice.
Hypertension research : official journal of the Japanese Society of Hypertension
Bi HL,Zhang YL,Yang J,Shu Q,Yang XL,Yan X,Chen C,Li Z,Li HH
Sun Mar 01 00:00:00 EST 2020
The Capsid Protein of Semliki Forest Virus Antagonizes RNA Interference in Mammalian Cells.
Journal of virology
Qian Q,Zhou H,Shu T,Mu J,Fang Y,Xu J,Li T,Kong J,Qiu Y,Zhou X
Fri Jan 17 00:00:00 EST 2020
The RCAN1.4-calcineurin/NFAT signaling pathway is essential for hypoxic adaption of intervertebral discs.
Experimental & molecular medicine
Huang B,He Y,Li S,Wei X,Liu J,Shan Z,Huang Y,Chen J,Zhao F
Fri May 01 00:00:00 EDT 2020
Knockdown of Rab9 Suppresses the Progression of Gastric Cancer Through Regulation of Akt Signaling Pathway.
Technology in cancer research & treatment
Zhu Y,Shi F,Wang M,Ding J
Fri Nov 20 00:00:00 EST 2020
Overexpression of KIAA0101 Promotes the Progression of Non-small Cell Lung Cancer.
Journal of Cancer
Cao H,Zheng J,Yao Y,Yang Q,Yan R,Sun W,Ruan K,Zhou J,Zhou J
Fri Oct 16 00:00:00 EDT 2020
Endogenous production of C-C motif chemokine ligand 2 by nasopharyngeal carcinoma cells drives radioresistance-associated metastasis.
Cancer letters
Guo SS,Liu R,Wen YF,Liu LT,Yuan L,Li YX,Li Y,Hao WW,Peng JY,Chen DN,Tang QN,Sun XS,Guo L,Mo HY,Qian CN,Zeng MS,Bei JX,Sun SY,Chen QY,Tang LQ,Mai HQ
Wed Jan 01 00:00:00 EST 2020
Sirtuin 2 Inhibition Attenuates Sevoflurane-Induced Learning and Memory Deficits in Developing Rats via Modulating Microglial Activation.
Cellular and molecular neurobiology
Wu Z,Zhang Y,Zhang Y,Zhao P
Wed Apr 01 00:00:00 EDT 2020
SYPL1 Inhibits Apoptosis in Pancreatic Ductal Adenocarcinoma via Suppression of ROS-Induced ERK Activation.
Frontiers in oncology
Song Y,Sun X,Duan F,He C,Wu J,Huang X,Xing K,Sun S,Wang R,Xie F,Mao Y,Wang J,Li S
Tue Oct 13 00:00:00 EDT 2020
Sema7A, a brain immune regulator, regulates seizure activity in PTZ-kindled epileptic rats.
CNS neuroscience & therapeutics
Deng J,Xu T,Yang J,Zhang KM,Li Q,Yu XY,Li R,Fu J,Jiang Q,Ma JX,Chen YM
Wed Jan 01 00:00:00 EST 2020
WDR62 is a novel participator in spindle migration and asymmetric cytokinesis during mouse oocyte meiotic maturation.
Experimental cell research
Wang YS,Jiao XF,Chen F,Wu D,Ding ZM,Miao YL,Huo LJ
Sat Feb 01 00:00:00 EST 2020
Paired box 9 regulates VSMC phenotypic transformation, proliferation, and migration via sonic hedgehog.
Life sciences
Xu J,Zhang Y,You S,Guo Y,Chen S,Chang Y,Zhang N,Sun Y
Tue Sep 15 00:00:00 EDT 2020
RNA-seq analysis of the key long noncoding RNAs and mRNAs related to cognitive impairment after cardiac arrest and cardiopulmonary resuscitation.
Aging
Chen C,Liu C,Niu Z,Li M,Zhang Y,Gao R,Chen H,Wang Q,Zhang S,Zhou R,Gan L,Zhang Z,Zhu T,Yu H,Liu J
Tue Jul 21 00:00:00 EDT 2020
MicroRNA-547-5p-mediated interleukin-33/suppressor of tumorigenicity 2 signaling underlies the genesis and maintenance of neuropathic pain and is targeted by the therapy with bone marrow stromal cells.
Molecular pain
Zhou J,Zhuang T,Ma P,Shan L,Sun XD,Gong S,Tao J,Yu XM,Jiang X
Thu Jun 03 00:00:00 EDT 2021
Inhibitory Activity of a Scorpion Defensin BmKDfsin3 against Hepatitis C Virus.
Antibiotics (Basel, Switzerland)
Cheng Y,Sun F,Li S,Gao M,Wang L,Sarhan M,Abdel-Rahman MA,Li W,Kwok HF,Wu Y,Cao Z
Fri Jan 17 00:00:00 EST 2020
Poly (ADP-ribosylation) of HMGB1 facilitates its acetylation and promotes HMGB1 translocation-associated chemotherapy-induced autophagy in leukaemia cells.
Oncology letters
Li Y,Xie J,Li X,Fang J
Wed Jan 01 00:00:00 EST 2020
Corticosterone Induced the Increase of proBDNF in Primary Hippocampal Neurons Via Endoplasmic Reticulum Stress.
Neurotoxicity research
Liu Y,Zou GJ,Tu BX,Hu ZL,Luo C,Cui YH,Xu Y,Li F,Dai RP,Bi FF,Li CQ
Sat Aug 01 00:00:00 EDT 2020
Regulation of Female Folliculogenesis by Tsp1a in Nile Tilapia (Oreochromis niloticus).
International journal of molecular sciences
Jie M,Ma H,Zhou L,Wu J,Li M,Liu X,Wang D
Sun Aug 16 00:00:00 EDT 2020
The chromatin remodeler Brg1 is required for formation and maintenance of hematopoietic stem cells.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Tu J,Liu X,Jia H,Reilly J,Yu S,Cai C,Liu F,Lv Y,Huang Y,Lu Z,Han S,Jiang T,Shu X,Wu X,Tang Z,Lu Q,Liu M
Tue Sep 01 00:00:00 EDT 2020
Acetylharpagide Protects Mice from Staphylococcus Aureus-Induced Acute Lung Injury by Inhibiting NF-κB Signaling Pathway.
Molecules (Basel, Switzerland)
Zhang Z,Wang Y,Shan Y,Yin W
Wed Nov 25 00:00:00 EST 2020
Macrophage-stimulating activity of European eel (Anguilla anguilla) peptides in RAW264.7 cells mediated via NF-κB and MAPK signaling pathways.
Food & function
Yao L,Yang P,Luo W,Li S,Wu Y,Cai N,Bi D,Li H,Han Q,Xu X
Tue Dec 01 00:00:00 EST 2020
Brain-Specific Serine-47 Modification of Cytochrome c Regulates Cytochrome c Oxidase Activity Attenuating ROS Production and Cell Death: Implications for Ischemia/Reperfusion Injury and Akt Signaling.
Cells
Kalpage HA,Wan J,Morse PT,Lee I,Hüttemann M
Thu Aug 06 00:00:00 EDT 2020
Functional analysis of natural PCSK9 mutants in modern and archaic humans.
The FEBS journal
Mikaeeli S,Susan-Resiga D,Girard E,Ben Djoudi Ouadda A,Day R,Prost S,Seidah NG
Sat Feb 01 00:00:00 EST 2020
Biallelic mutation of HSD17B4 induces middle age-onset spinocerebellar ataxia.
Neurology. Genetics
Matsuda Y,Morino H,Miyamoto R,Kurashige T,Kume K,Mizuno N,Kanaya Y,Tada Y,Ohsawa R,Yokota K,Shimozawa N,Maruyama H,Kawakami H
Sat Feb 01 00:00:00 EST 2020
Disregulation of Autophagy in the Transgenerational Cc2d1a Mouse Model of Autism.
Neuromolecular medicine
Dana H,Bayramov KK,Delibaşı N,Tahtasakal R,Bayramov R,Hamurcu Z,Sener EF
Mon Jun 01 00:00:00 EDT 2020
RNF126-Mediated Reubiquitination Is Required for Proteasomal Degradation of p97-Extracted Membrane Proteins.
Molecular cell
Hu X,Wang L,Wang Y,Ji J,Li J,Wang Z,Li C,Zhang Y,Zhang ZR
Thu Jul 16 00:00:00 EDT 2020
Pum2 mediates Sirt1 mRNA decay and exacerbates hypoxia/reoxygenation-induced cardiomyocyte apoptosis.
Experimental cell research
Cao Y,Liu C,Wang Q,Wang W,Tao E,Wan L
Sat Aug 01 00:00:00 EDT 2020
Metformin improves boar sperm quality via 5'-AMP-activated protein kinase-mediated energy metabolism in vitro.
Zoological research
Li RN,Zhu ZD,Zheng Y,Lv YH,Tian XE,Wu D,Wang YJ,Zeng WX
Fri Sep 18 00:00:00 EDT 2020
NQO1-selective activated prodrugs of combretastatin A-4: Synthesis and biological evaluation.
Bioorganic chemistry
Zhang C,Qu Y,Ma X,Li M,Li S,Li Y,Wu L
Thu Oct 01 00:00:00 EDT 2020
SIRT6-PARP1 is involved in HMGB1 polyADP-ribosylation and acetylation and promotes chemotherapy-induced autophagy in leukemia.
Cancer biology & therapy
Kong Q,Li Y,Liang Q,Xie J,Li X,Fang J
Thu Apr 02 00:00:00 EDT 2020
PHLDA1 is a new therapeutic target of oxidative stress and ischemia reperfusion-induced myocardial injury.
Life sciences
Guo Y,Jia P,Chen Y,Yu H,Xin X,Bao Y,Yang H,Wu N,Sun Y,Jia D
Sun Mar 15 00:00:00 EDT 2020
Wnt5a/CaMKII/ERK/CCL2 axis is required for tumor-associated macrophages to promote colorectal cancer progression.
International journal of biological sciences
Liu Q,Song J,Pan Y,Shi D,Yang C,Wang S,Xiong B
Mon Jun 07 00:00:00 EDT 2021
Inward Tension of Talin and Integrin-related Osmotic Pressure are involved Synergetically in the Invasion and Metastasis of Non-small Cell Lung Cancer.
Journal of Cancer
Song Y,Li C,Fu Y,Xie Q,Guo J,Li G,Wu H
Mon Sep 28 00:00:00 EDT 2020
A Prognostic Prediction Model Developed Based on Four CpG Sites and Weighted Correlation Network Analysis Identified DNAJB1 as a Novel Biomarker for Pancreatic Cancer.
Frontiers in oncology
Kong L,Liu P,Fei X,Wu T,Wang Z,Zhang B,Li J,Tan X
Sun Apr 17 00:00:00 EDT 2022
Broad-complex transcription factor mediates opposing hormonal regulation of two phylogenetically distant arginine kinase genes in Tribolium castaneum.
Communications biology
Zhang N,Jiang H,Meng X,Qian K,Liu Y,Song Q,Stanley D,Wu J,Park Y,Wang J
Fri Oct 30 00:00:00 EDT 2020
Oroxylin A alleviates immunoparalysis of CLP mice by degrading CHOP through interacting with FBXO15.
Scientific reports
Zhang Z,Wang Y,Shan Y,Zhou R,Yin W
Fri Nov 06 00:00:00 EST 2020
Aster-B coordinates with Arf1 to regulate mitochondrial cholesterol transport.
Molecular metabolism
Andersen JP,Zhang J,Sun H,Liu X,Liu J,Nie J,Shi Y
Tue Dec 01 00:00:00 EST 2020
Hepatic CD147 knockout modulates liver steatosis and up-regulates autophagy in high-fat-diet-induced NAFLD mice.
Biochemical and biophysical research communications
Lou J,Li C,Li ZS,Zhang T,Chen ZN,Bian H
Thu Apr 16 00:00:00 EDT 2020
Glutamine protects intestine against ischemia-reperfusion injury by alleviating endoplasmic reticulum stress induced apoptosis in rats.
Acta cirurgica brasileira
Xu H,Liu G,Gu H,Wang J,Li Y
Mon Mar 23 00:00:00 EDT 2020
Super-enhancer-driven metabolic reprogramming promotes cystogenesis in autosomal dominant polycystic kidney disease.
Nature metabolism
Mi Z,Song Y,Cao X,Lu Y,Liu Z,Zhu X,Geng M,Sun Y,Lan B,He C,Xiong H,Zhang L,Chen Y
Sat Aug 01 00:00:00 EDT 2020
HOXA13 promotes colon cancer progression through β-catenin-dependent WNT pathway.
Experimental cell research
Gu Y,Gu J,Shen K,Zhou H,Hao J,Li F,Yu H,Chen Y,Li J,Li Y,Liang H,Dong Y
Thu Oct 15 00:00:00 EDT 2020
Bnip3 mediates doxorubicin-induced cardiomyocyte pyroptosis via caspase-3/GSDME.
Life sciences
Zheng X,Zhong T,Ma Y,Wan X,Qin A,Yao B,Zou H,Song Y,Yin D
Sat Feb 01 00:00:00 EST 2020
Comprehensive lipidomics in apoM-/- mice reveals an overall state of metabolic distress and attenuated hepatic lipid secretion into the circulation.
Journal of genetics and genomics = Yi chuan xue bao
Shi Y,Lam SM,Liu H,Luo G,Zhang J,Yao S,Li J,Zheng L,Xu N,Zhang X,Shui G
Sun Sep 20 00:00:00 EDT 2020
Proline Protects Boar Sperm against Oxidative Stress through Proline Dehydrogenase-Mediated Metabolism and the Amine Structure of Pyrrolidine.
Animals : an open access journal from MDPI
Feng C,Zhu Z,Bai W,Li R,Zheng Y,Tian X,Wu D,Lu H,Wang Y,Zeng W
Tue Sep 01 00:00:00 EDT 2020
MicroRNA-148a regulates low-density lipoprotein metabolism by repressing the (pro)renin receptor.
PloS one
Wang N,He L,Lin H,Tan L,Sun Y,Zhang X,Danser AHJ,Lu HS,He Y,Lu X
Wed Sep 09 00:00:00 EDT 2020
Phosphoribosyl pyrophosphate synthetases 2 knockdown inhibits prostate cancer progression by suppressing cell cycle and inducing cell apoptosis.
Journal of Cancer
Qiao H,Tan X,Lv DJ,Xing RW,Shu FP,Zhong CF,Li C,Zou YG,Mao XM
Wed Sep 30 00:00:00 EDT 2020
Centrosome Amplification in Cancer Disrupts Autophagy and Sensitizes to Autophagy Inhibition.
Molecular cancer research : MCR
Denu RA,Kaur G,Sass MM,Lakkaraju A,Burkard ME
Wed Jan 01 00:00:00 EST 2020
Alternatively Polarized Macrophages Regulate the Growth and Differentiation of Ependymal Stem Cells through the SIRT2 Pathway.
Experimental neurobiology
Ma Y,Deng M,Zhao XQ,Liu M
Thu Apr 30 00:00:00 EDT 2020
Dlgap1 negatively regulates browning of white fat cells through effects on cell proliferation and apoptosis.
Lipids in health and disease
Zhang J,Yang J,Yang N,Ma J,Lu D,Dong Y,Liang H,Liu D,Cang M
Fri Mar 13 00:00:00 EDT 2020
Downregulated miR-150 in bone marrow mesenchymal stem cells attenuates the apoptosis of LPS-stimulated RAW264.7 via MTCH2-dependent mitochondria transfer.
Biochemical and biophysical research communications
Zhou X,Zhang K,He Z,Deng Y,Gao Y
Thu Jun 04 00:00:00 EDT 2020
Cytoplasmic PCNA is located in the actin belt and involved in osteoclast differentiation.
Aging
Tang D,Liu X,Chen K,Li Z,Dai Y,Xu J,Zhang HT,Gao X,Liu L
Sat Jun 27 00:00:00 EDT 2020
AFF1 inhibits adipogenic differentiation via targeting TGM2 transcription.
Cell proliferation
Chen Y,Wang Y,Lin W,Sheng R,Wu Y,Xu R,Zhou C,Yuan Q
Mon Jun 01 00:00:00 EDT 2020
Structural basis for RING-Cys-Relay E3 ligase activity and its role in axon integrity.
Nature chemical biology
Mabbitt PD,Loreto A,Déry MA,Fletcher AJ,Stanley M,Pao KC,Wood NT,Coleman MP,Virdee S
Sun Nov 01 00:00:00 EDT 2020
Lycorine hydrochloride inhibits cell proliferation and induces apoptosis through promoting FBXW7-MCL1 axis in gastric cancer.
Journal of experimental & clinical cancer research : CR
Li C,Deng C,Pan G,Wang X,Zhang K,Dong Z,Zhao G,Tan M,Hu X,Shi S,Du J,Ji H,Wang X,Yang L,Cui H
Fri Oct 30 00:00:00 EDT 2020
Atractyloside targets cancer-associated fibroblasts and inhibits the metastasis of colon cancer.
Annals of translational medicine
Qi L,Song F,Han Y,Zhang Y,Ding Y
Sun Nov 01 00:00:00 EDT 2020
Novel poricoic acids attenuate renal fibrosis through regulating redox signalling and aryl hydrocarbon receptor activation.
Phytomedicine : international journal of phytotherapy and phytopharmacology
Wang M,Hu HH,Chen YY,Chen L,Wu XQ,Zhao YY
Tue Dec 01 00:00:00 EST 2020
Fine particulate matter increases airway hyperresponsiveness through kallikrein-bradykinin pathway.
Ecotoxicology and environmental safety
Cao X,Wang M,Li J,Luo Y,Li R,Yan X,Zhang H
Mon Jun 01 00:00:00 EDT 2020
AIM2 promotes the development of non-small cell lung cancer by modulating mitochondrial dynamics.
Oncogene
Qi M,Dai D,Liu J,Li Z,Liang P,Wang Y,Cheng L,Zhan Y,An Z,Song Y,Yang Y,Yan X,Xiao H,Shao H
Sun Mar 01 00:00:00 EST 2020
Herb-sourced emodin inhibits angiogenesis of breast cancer by targeting VEGFA transcription.
Theranostics
Zou G,Zhang X,Wang L,Li X,Xie T,Zhao J,Yan J,Wang L,Ye H,Jiao S,Xiang R,Shi Y
Tue May 18 00:00:00 EDT 2021
An MRTF-A-Sp1-PDE5 Axis Mediates Angiotensin-II-Induced Cardiomyocyte Hypertrophy.
Frontiers in cell and developmental biology
Wu T,Wang H,Xin X,Yang J,Hou Y,Fang M,Lu X,Xu Y
Fri Jul 16 00:00:00 EDT 2021
Cell Signaling Coordinates Global PRC2 Recruitment and Developmental Gene Expression in Murine Embryonic Stem Cells.
iScience
Aljazi MB,Gao Y,Wu Y,Mias GI,He J
Fri Nov 20 00:00:00 EST 2020
Mogroside V Protects against Hepatic Steatosis in Mice on a High-Fat Diet and LO2 Cells Treated with Free Fatty Acids via AMPK Activation.
Evidence-based complementary and alternative medicine : eCAM
Li L,Zheng W,Wang C,Qi J,Li H
Mon Sep 28 00:00:00 EDT 2020
Interleukin-17 suppresses grass carp reovirus infection in Ctenopharyngodon idellus kidney cells by activating NF-κB signaling.
Aquaculture (Amsterdam, Netherlands)
Zhang Y,Zhang X,Liang Z,Dai K,Zhu M,Zhang M,Pan J,Xue R,Cao G,Tang J,Song X,Hu X,Gong C
Wed Apr 15 00:00:00 EDT 2020
Coxsackievirus A6 Induces Necroptosis for Viral Production.
Frontiers in microbiology
Zhang S,Yu X,Meng X,Huo W,Su Y,Liu J,Liu Y,Zhang J,Wang S,Yu J
Mon Sep 28 00:00:00 EDT 2020
Cognitive improvement and synaptic deficit attenuation by a multifunctional carbazole-based cyanine in AD mice model through regulation of Ca2+/CaMKII/CREB signaling pathway.
Experimental neurology
Chen C,Xu D,Zhang ZH,Jia SZ,Cao XC,Chen YB,Song GL,Wong MS,Li HW
Fri May 01 00:00:00 EDT 2020
Melatonin Rescues the Ti Particle-Impaired Osteogenic Potential of Bone Marrow Mesenchymal Stem Cells via the SIRT1/SOD2 Signaling Pathway.
Calcified tissue international
Zhang Y,Zhu X,Wang G,Chen L,Yang H,He F,Lin J
Sun Nov 01 00:00:00 EDT 2020
Downregulation of the circadian rhythm regulator HLF promotes multiple-organ distant metastases in non-small cell lung cancer through PPAR/NF-κb signaling.
Cancer letters
Chen J,Liu A,Lin Z,Wang B,Chai X,Chen S,Lu W,Zheng M,Cao T,Zhong M,Li R,Wu M,Lu Z,Pang W,Huang W,Xiao L,Lin D,Wang Z,Lei F,Chen X,Long W,Zheng Y,Chen Q,Zeng J,Ren D,Li J,Zhang X,Huang Y
Fri Jul 10 00:00:00 EDT 2020
Expanding the retinal phenotype of RP1: from retinitis pigmentosa to a novel and singular macular dystrophy.
The British journal of ophthalmology
Riera M,Abad-Morales V,Navarro R,Ruiz-Nogales S,Méndez-Vendrell P,Corcostegui B,Pomares E
Sat Feb 01 00:00:00 EST 2020
Soluble Uric Acid Promotes Atherosclerosis via AMPK (AMP-Activated Protein Kinase)-Mediated Inflammation.
Arteriosclerosis, thrombosis, and vascular biology
Kimura Y,Yanagida T,Onda A,Tsukui D,Hosoyamada M,Kono H
Sun Mar 01 00:00:00 EST 2020
Synthesis and biological evaluation of (3/4-(pyrimidin-2-ylamino)benzoyl)-based hydrazine-1-carboxamide/carbothioamide derivatives as novel RXRα antagonists.
Journal of enzyme inhibition and medicinal chemistry
Qin J,Liu J,Wu C,Xu J,Tang B,Guo K,Chen X,Liu W,Wu T,Zhou H,Fang M,Wu Z
Tue Dec 01 00:00:00 EST 2020
Elevated miR-124-3p in the aging colon disrupts mucus barrier and increases susceptibility to colitis by targeting T-synthase.
Aging cell
Huang L,Sun TY,Hu LJ,Hu SL,Sun HM,Zhao FQ,Wu B,Yang S,Ji FQ,Zhou DS
Sun Nov 01 00:00:00 EDT 2020
Immune activation of murine RAW264.7 macrophages by sonicated and alkalized paramylon from Euglena gracilis.
BMC microbiology
Guo Q,Bi D,Wu M,Yu B,Hu L,Liu C,Gu L,Zhu H,Lei A,Xu X,Wang J
Fri Jun 19 00:00:00 EDT 2020
MACF1 promotes preosteoblast migration by mediating focal adhesion turnover through EB1.
Biology open
Su P,Yin C,Li D,Yang C,Wang X,Pei J,Tian Y,Qian A
Tue Mar 24 00:00:00 EDT 2020
Plakoglobin is involved in cytoskeletal rearrangement of podocytes under the regulation of UCH-L1.
Biochemical and biophysical research communications
Fang Y,Li F,Qi C,Mao X,Xu Y,Zhao Z,Wu H,Zhang Z
Thu Aug 13 00:00:00 EDT 2020
CXCL12/CXCR4 signaling induced itch and pain sensation in a murine model of allergic contact dermatitis.
Molecular pain
Su W,Yu J,Liu Q,Ma L,Huang Y
Thu Jun 03 00:00:00 EDT 2021
Ubiquitin C-terminal hydrolase L1 (UCHL1) regulates post-myocardial infarction cardiac fibrosis through glucose-regulated protein of 78 kDa (GRP78).
Scientific reports
Lei Q,Yi T,Li H,Yan Z,Lv Z,Li G,Wang Y
Tue Jun 30 00:00:00 EDT 2020
The nuclear translocation of transketolase inhibits the farnesoid receptor expression by promoting the binding of HDAC3 to FXR promoter in hepatocellular carcinoma cell lines.
Cell death & disease
Li M,Zhang X,Lu Y,Meng S,Quan H,Hou P,Tong P,Chai D,Gao X,Zheng J,Tong X,Bai J
Thu Jan 16 00:00:00 EST 2020
Arginase 1 is involved in lacrimal hyposecretion in male NOD mice, a model of Sjögren's syndrome, regardless of dacryoadenitis status.
The Journal of physiology
Ohno Y,Satoh K,Shitara A,Into T,Kashimata M
Sun Nov 01 00:00:00 EDT 2020
ZNF582 hypermethylation promotes metastasis of nasopharyngeal carcinoma by regulating the transcription of adhesion molecules Nectin-3 and NRXN3.
Cancer communications (London, England)
Zhao Y,Hong XH,Li K,Li YQ,Li YQ,He SW,Zhang PP,Li JY,Li Q,Liang YL,Chen Y,Ma J,Liu N,Chen YP
Tue Dec 01 00:00:00 EST 2020
Anti-inflammatory and analgesic activities of indigo through regulating the IKKβ/IκB/NF-κB pathway in mice.
Food & function
Liu N,Zhang GX,Niu YT,Wang Q,Zheng J,Yang JM,Sun T,Niu JG,Yu JQ
Thu Oct 01 00:00:00 EDT 2020
Expression and significance of T-cell immunoglobulin mucin molecule 3 and its ligand galectin-9 in patients with adenomyosis.
Gynecological endocrinology : the official journal of the International Society of Gynecological Endocrinology
Huang P,Lv C,Zhang C,Feng H,Li C,Zhang H,Zhao X,Li M
Wed Jul 01 00:00:00 EDT 2020
lncRNA SLC7A11-AS1 Promotes Chemoresistance by Blocking SCFβ-TRCP-Mediated Degradation of NRF2 in Pancreatic Cancer.
Molecular therapy. Nucleic acids
Yang Q,Li K,Huang X,Zhao C,Mei Y,Li X,Jiao L,Yang H
Fri Mar 06 00:00:00 EST 2020
EBV-miR-BART12 accelerates migration and invasion in EBV-associated cancer cells by targeting tubulin polymerization-promoting protein 1.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Wu Y,Wang D,Wei F,Xiong F,Zhang S,Gong Z,Shi L,Li X,Xiang B,Ma J,Deng H,He Y,Liao Q,Zhang W,Li X,Li Y,Guo C,Zeng Z,Li G,Xiong W
Tue Dec 01 00:00:00 EST 2020
Betulinic Acid-Mediated Tuning of PERK/CHOP Signaling by Sp1 Inhibition as a Novel Therapeutic Strategy for Glioblastoma.
Cancers
Lo WL,Hsu TI,Yang WB,Kao TJ,Wu MH,Huang YN,Yeh SH,Chuang JY
Wed Apr 15 00:00:00 EDT 2020
Caspase-3 inhibitor inhibits enterovirus D68 production.
Journal of microbiology (Seoul, Korea)
Huo W,Yu J,Liu C,Wu T,Wang Y,Meng X,Song F,Zhang S,Su Y,Liu Y,Liu J,Yu X,Hua S
Tue Sep 01 00:00:00 EDT 2020
Krüppel-like factor 3 inhibition by mutated lncRNA Reg1cp results in human high bone mass syndrome.
The Journal of experimental medicine
Yang M,Guo Q,Peng H,Xiao YZ,Xiao Y,Huang Y,Li CJ,Su T,Zhang YL,Lei MX,Chen HL,Jiang TJ,Luo XH
Mon Aug 05 00:00:00 EDT 2019
AMPK Promotes SPOP-Mediated NANOG Degradation to Regulate Prostate Cancer Cell Stemness.
Developmental cell
Wang X,Jin J,Wan F,Zhao L,Chu H,Chen C,Liao G,Liu J,Yu Y,Teng H,Fang L,Jiang C,Pan W,Xie X,Li J,Lu X,Jiang X,Ge X,Ye D,Wang P
Mon Feb 11 00:00:00 EST 2019
AMPK Promotes SPOP-Mediated NANOG Degradation to Regulate Prostate Cancer Cell Stemness.
Developmental cell
Wang X,Jin J,Wan F,Zhao L,Chu H,Chen C,Liao G,Liu J,Yu Y,Teng H,Fang L,Jiang C,Pan W,Xie X,Li J,Lu X,Jiang X,Ge X,Ye D,Wang P
Mon Feb 11 00:00:00 EST 2019
CERKL regulates autophagy via the NAD-dependent deacetylase SIRT1.
Autophagy
Hu X,Lu Z,Yu S,Reilly J,Liu F,Jia D,Qin Y,Han S,Liu X,Qu Z,Lv Y,Li J,Huang Y,Jiang T,Jia H,Wang Q,Liu J,Shu X,Tang Z,Liu M
Fri Mar 01 00:00:00 EST 2019
CERKL regulates autophagy via the NAD-dependent deacetylase SIRT1.
Autophagy
Hu X,Lu Z,Yu S,Reilly J,Liu F,Jia D,Qin Y,Han S,Liu X,Qu Z,Lv Y,Li J,Huang Y,Jiang T,Jia H,Wang Q,Liu J,Shu X,Tang Z,Liu M
Fri Mar 01 00:00:00 EST 2019
CERKL regulates autophagy via the NAD-dependent deacetylase SIRT1.
Autophagy
Hu X,Lu Z,Yu S,Reilly J,Liu F,Jia D,Qin Y,Han S,Liu X,Qu Z,Lv Y,Li J,Huang Y,Jiang T,Jia H,Wang Q,Liu J,Shu X,Tang Z,Liu M
Fri Mar 01 00:00:00 EST 2019
Interferon-alpha promotes immunosuppression through IFNAR1/STAT1 signalling in head and neck squamous cell carcinoma.
British journal of cancer
Ma H,Yang W,Zhang L,Liu S,Zhao M,Zhou G,Wang L,Jin S,Zhang Z,Hu J
Fri Feb 01 00:00:00 EST 2019
Interferon-alpha promotes immunosuppression through IFNAR1/STAT1 signalling in head and neck squamous cell carcinoma.
British journal of cancer
Ma H,Yang W,Zhang L,Liu S,Zhao M,Zhou G,Wang L,Jin S,Zhang Z,Hu J
Fri Feb 01 00:00:00 EST 2019
MSC-regulated lncRNA MACC1-AS1 promotes stemness and chemoresistance through fatty acid oxidation in gastric cancer.
Oncogene
He W,Liang B,Wang C,Li S,Zhao Y,Huang Q,Liu Z,Yao Z,Wu Q,Liao W,Zhang S,Liu Y,Xiang Y,Liu J,Shi M
Sat Jun 01 00:00:00 EDT 2019
Myocardial Adipose Triglyceride Lipase Overexpression Protects against Burn-Induced Cardiac Lipid Accumulation and Injury.
Oxidative medicine and cellular longevity
Li L,Zhang X,Zhang Q,Jia J,Zhang J,Zhang D,Song H,Chen B,Hu J,Huang Y
Fri Dec 13 00:00:00 EST 2019
CD147 promotes progression of head and neck squamous cell carcinoma via NF-kappa B signaling.
Journal of cellular and molecular medicine
Yu B,Zhang Y,Wu K,Wang L,Jiang Y,Chen W,Yan M
Fri Feb 01 00:00:00 EST 2019
CD147 promotes progression of head and neck squamous cell carcinoma via NF-kappa B signaling.
Journal of cellular and molecular medicine
Yu B,Zhang Y,Wu K,Wang L,Jiang Y,Chen W,Yan M
Fri Feb 01 00:00:00 EST 2019
Altered expression of p63 isoforms and expansion of p63- and club cell secretory protein-positive epithelial cells in the lung as novel features of aging.
American journal of physiology. Cell physiology
Fukumoto J,Sidramagowda Patil S,Krishnamurthy S,Saji S,John I,Narala VR,Hernández-Cuervo H,Alleyn M,Breitzig MT,Galam L,Soundararajan R,Chaudhari UK,Hansen BC,Lockey RF,Kolliputi N
Mon Apr 01 00:00:00 EDT 2019
An increase of estrogen receptor α protein level regulates BDE-209-mediated blood-testis barrier disruption during spermatogenesis in F1 mice.
Environmental science and pollution research international
Zhai J,Geng X,Ding T,Li J,Tang J,Chen D,Cui L,Wang Q
Fri Feb 01 00:00:00 EST 2019
An increase of estrogen receptor α protein level regulates BDE-209-mediated blood-testis barrier disruption during spermatogenesis in F1 mice.
Environmental science and pollution research international
Zhai J,Geng X,Ding T,Li J,Tang J,Chen D,Cui L,Wang Q
Fri Feb 01 00:00:00 EST 2019
Lamin A mutation impairs interaction with nucleoporin NUP155 and disrupts nucleocytoplasmic transport in atrial fibrillation.
Human mutation
Han M,Zhao M,Cheng C,Huang Y,Han S,Li W,Tu X,Luo X,Yu X,Liu Y,Chen Q,Ren X,Wang QK,Ke T
Fri Mar 01 00:00:00 EST 2019
Lamin A mutation impairs interaction with nucleoporin NUP155 and disrupts nucleocytoplasmic transport in atrial fibrillation.
Human mutation
Han M,Zhao M,Cheng C,Huang Y,Han S,Li W,Tu X,Luo X,Yu X,Liu Y,Chen Q,Ren X,Wang QK,Ke T
Fri Mar 01 00:00:00 EST 2019
FOXO1 associated with sensitivity to chemotherapy drugs and glial-mesenchymal transition in glioma.
Journal of cellular biochemistry
Chen C,Han G,Li Y,Yue Z,Wang L,Liu J
Tue Jan 01 00:00:00 EST 2019
FOXO1 associated with sensitivity to chemotherapy drugs and glial-mesenchymal transition in glioma.
Journal of cellular biochemistry
Chen C,Han G,Li Y,Yue Z,Wang L,Liu J
Tue Jan 01 00:00:00 EST 2019
NPRL2 enhances autophagy and the resistance to Everolimus in castration-resistant prostate cancer.
The Prostate
Chen Z,Jiang Q,Zhu P,Chen Y,Xie X,Du Z,Jiang L,Tang W
Tue Jan 01 00:00:00 EST 2019
NPRL2 enhances autophagy and the resistance to Everolimus in castration-resistant prostate cancer.
The Prostate
Chen Z,Jiang Q,Zhu P,Chen Y,Xie X,Du Z,Jiang L,Tang W
Tue Jan 01 00:00:00 EST 2019
TGF‑β induces periodontal ligament stem cell senescence through increase of ROS production.
Molecular medicine reports
Fan C,Ji Q,Zhang C,Xu S,Sun H,Li Z
Tue Oct 01 00:00:00 EDT 2019
MiR-15b-5p is Involved in Doxorubicin-Induced Cardiotoxicity via Inhibiting Bmpr1a Signal in H9c2 Cardiomyocyte.
Cardiovascular toxicology
Wan GX,Cheng L,Qin HL,Zhang YZ,Wang LY,Zhang YG
Sat Jun 01 00:00:00 EDT 2019
MiR-15b-5p is Involved in Doxorubicin-Induced Cardiotoxicity via Inhibiting Bmpr1a Signal in H9c2 Cardiomyocyte.
Cardiovascular toxicology
Wan GX,Cheng L,Qin HL,Zhang YZ,Wang LY,Zhang YG
Sat Jun 01 00:00:00 EDT 2019
The LIV-1-GRPEL1 axis adjusts cell fate during anti-mitotic agent-damaged mitosis.
EBioMedicine
Chen P,Wang B,Mo Q,Wu P,Fang Y,Tian Y,Jin X,Gao Y,Wu Y,Cao Y,Zhang Y,Xi L,Wang S,Hu J,Ma D,Zhou J,Gao Q,Chen G
Fri Nov 01 00:00:00 EDT 2019
VEGF Induce Vasculogenic Mimicry of Choroidal Melanoma through the PI3k Signal Pathway.
BioMed research international
Xu X,Zong Y,Gao Y,Sun X,Zhao H,Luo W,Jia S
Thu Jan 02 00:00:00 EST 2020
Fine-Tuning of Shh/Gli Signaling Gradient by Non-proteolytic Ubiquitination during Neural Patterning.
Cell reports
Ma P,Song NN,Li Y,Zhang Q,Zhang L,Zhang L,Kong Q,Ma L,Yang X,Ren B,Li C,Zhao X,Li Y,Xu Y,Gao X,Ding YQ,Mao B
Tue Jul 09 00:00:00 EDT 2019
Helicobacter pylori induces intracellular galectin-8 aggregation around damaged lysosomes within gastric epithelial cells in a host O-glycan-dependent manner.
Glycobiology
Li FY,Weng IC,Lin CH,Kao MC,Wu MS,Chen HY,Liu FT
Fri Feb 01 00:00:00 EST 2019
The Mechanism of Rap1 Regulates N-cadherin to Control Neuronal Migration.
Journal of molecular neuroscience : MN
Yang C,Li X,Zhang B,Fu S,Li S,Shen J,Guan L,Qiao L,Lin J
Thu Aug 01 00:00:00 EDT 2019
Proteomics Profiling of Host Cell Response via Protein Expression and Phosphorylation upon Dengue Virus Infection.
Virologica Sinica
Miao M,Yu F,Wang D,Tong Y,Yang L,Xu J,Qiu Y,Zhou X,Zhao X
Tue Oct 01 00:00:00 EDT 2019
NFAT1 Hypermethylation Promotes Epithelial-Mesenchymal Transition and Metastasis in Nasopharyngeal Carcinoma by Activating ITGA6 Transcription.
Neoplasia (New York, N.Y.)
Zhang J,Zheng ZQ,Yuan YW,Zhang PP,Li YQ,Wang YQ,Tang XR,Wen X,Hong XH,Lei Y,He QM,Yang XJ,Sun Y,Ma J,Liu N
Fri Mar 01 00:00:00 EST 2019
Accumulation of fructose 1,6-bisphosphate protects clear cell renal cell carcinoma from oxidative stress.
Laboratory investigation; a journal of technical methods and pathology
Wang J,Wu Q,Qiu J
Sat Jun 01 00:00:00 EDT 2019
Targeting the mDia Formin-Assembled Cytoskeleton Is an Effective Anti-Invasion Strategy in Adult High-Grade Glioma Patient-Derived Neurospheres.
Cancers
Pettee KM,Becker KN,Alberts AS,Reinard KA,Schroeder JL,Eisenmann KM
Wed Mar 20 00:00:00 EDT 2019
Qingpeng Ointment Ameliorates Inflammatory Responses and Dysregulation of Itch-Related Molecules for Its Antipruritic Effects in Experimental Allergic Contact Dermatitis.
Frontiers in pharmacology
Gong X,Xiong H,Liu S,Liu Y,Yin L,Tu C,Wang H,Zhao Z,Chen W,Mei Z
Thu Oct 01 00:00:00 EDT 2020
Inhibition of PI3K/mTOR increased the sensitivity of hepatocellular carcinoma cells to cisplatin via interference with mitochondrial-lysosomal crosstalk.
Cell proliferation
Sheng J,Shen L,Sun L,Zhang X,Cui R,Wang L
Wed May 01 00:00:00 EDT 2019
Antitumor effects and mechanisms of 1,25(OH)2D3 in the Pfeiffer diffuse large B lymphoma cell line.
Molecular medicine reports
Han J,Tang Y,Zhong M,Wu W
Sun Dec 01 00:00:00 EST 2019
Ovarian cancer cell-secreted exosomal miR-205 promotes metastasis by inducing angiogenesis.
Theranostics
He L,Zhu W,Chen Q,Yuan Y,Wang Y,Wang J,Wu X
Fri Aug 28 00:00:00 EDT 2020
The histone demethylase Kdm4 suppresses activation of hepatic stellate cell by inducing MiR-29 transcription.
Biochemical and biophysical research communications
Kong M,Wu J,Fan Z,Chen B,Wu T,Xu Y
Tue Jun 18 00:00:00 EDT 2019
STRIP2 silencing inhibits vascular smooth muscle cell proliferation and migration via P38-AKT-MMP-2 signaling pathway.
Journal of cellular physiology
Dai L,Zhou J,Li T,Qian Y,Jin L,Zhu C,Li S
Sun Dec 01 00:00:00 EST 2019
Large-conductance calcium-activated potassium channels mediate lipopolysaccharide-induced activation of murine microglia.
The Journal of biological chemistry
Yang X,Wang G,Cao T,Zhang L,Ma Y,Jiang S,Teng X,Sun X
Fri Aug 30 00:00:00 EDT 2019
The pivotal role of protein acetylation in linking glucose and fatty acid metabolism to β-cell function.
Cell death & disease
Zhang Y,Zhou F,Bai M,Liu Y,Zhang L,Zhu Q,Bi Y,Ning G,Zhou L,Wang X
Fri Jan 25 00:00:00 EST 2019
Inactive USP14 and inactive UCHL5 cause accumulation of distinct ubiquitinated proteins in mammalian cells.
PloS one
Chadchankar J,Korboukh V,Conway LC,Wobst HJ,Walker CA,Doig P,Jacobsen SJ,Brandon NJ,Moss SJ,Wang Q
Mon Mar 23 00:00:00 EDT 2020
Mitochondrial pyruvate carrier 2 mediates mitochondrial dysfunction and apoptosis in high glucose-treated podocytes.
Life sciences
Feng J,Ma Y,Chen Z,Hu J,Yang Q,Ding G
Fri Nov 15 00:00:00 EST 2019
Membrane association but not identity is required for LRRK2 activation and phosphorylation of Rab GTPases.
The Journal of cell biology
Gomez RC,Wawro P,Lis P,Alessi DR,Pfeffer SR
Mon Dec 02 00:00:00 EST 2019
Cul4a as a New Interaction Protein of PARP1 Inhibits Oxidative Stress-Induced H9c2 Cell Apoptosis.
Oxidative medicine and cellular longevity
Ye N,Zhang N,Zhang Y,Qian H,Wu B,Sun Y
Thu Jan 16 00:00:00 EST 2020
Design, synthesis, antitumor activities and biological studies of novel diaryl substituted fused heterocycles as dual ligands targeting tubulin and katanin.
European journal of medicinal chemistry
Gao F,Liang Y,Zhou P,Cheng J,Ding K,Wang Y
Sun Sep 15 00:00:00 EDT 2019
Effects of Betaine on LPS-Stimulated Activation of Microglial M1/M2 Phenotypes by Suppressing TLR4/NF-κB Pathways in N9 Cells.
Molecules (Basel, Switzerland)
Shi H,Wang XL,Quan HF,Yan L,Pei XY,Wang R,Peng XD
Mon Jan 21 00:00:00 EST 2019
DDX23, an Evolutionary Conserved dsRNA Sensor, Participates in Innate Antiviral Responses by Pairing With TRIF or MAVS.
Frontiers in immunology
Ruan J,Cao Y,Ling T,Li P,Wu S,Peng D,Wang Y,Jia X,Chen S,Xu A,Yuan S
Tue Nov 10 00:00:00 EST 2020
Inhibition of glycogen catabolism induces intrinsic apoptosis and augments multikinase inhibitors in hepatocellular carcinoma cells.
Experimental cell research
Barot S,Abo-Ali EM,Zhou DL,Palaguachi C,Dukhande VV
Thu Aug 15 00:00:00 EDT 2019
Osteoprotegerin promotes islet β cell proliferation in intrauterine growth retardation rats through the PI3K/AKT/FoxO1 pathway.
International journal of clinical and experimental pathology
Tang S,Xin Y,Yang M,Zhang D,Xu C
Fri Jan 17 00:00:00 EST 2020
Enhancement of cortisol-induced SAA1 transcription by SAA1 in the human amnion.
Journal of molecular endocrinology
Lu Y,Wang WS,Lin YK,Lu JW,Li WJ,Zhang CY,Sun K
Wed May 01 00:00:00 EDT 2019
MicroRNA-125a-3p affects smooth muscle cell function in vascular stenosis.
Journal of molecular and cellular cardiology
Hu W,Chang G,Zhang M,Li Y,Yin L,Huang Y,Feng C,Gu Y,Wen D,Wang S
Fri Nov 01 00:00:00 EDT 2019
HMGB1 and Caveolin-1 related to RPE cell senescence in age-related macular degeneration.
Aging
Sun S,Cai B,Li Y,Su W,Zhao X,Gong B,Li Z,Zhang X,Wu Y,Chen C,Tsang SH,Yang J,Li X
Sun Jul 07 00:00:00 EDT 2019
The Novel Nature Microtubule Inhibitor Ivalin Induces G2/M Arrest and Apoptosis in Human Hepatocellular Carcinoma SMMC-7721 Cells In Vitro.
Medicina (Kaunas, Lithuania)
Liu F,Lin S,Zhang C,Ma J,Han Z,Jia F,Xie W,Li X
Mon Aug 12 00:00:00 EDT 2019
Identification of the Potential Key Long Non-coding RNAs in Aged Mice With Postoperative Cognitive Dysfunction.
Frontiers in aging neuroscience
Li M,Chen C,Zhang W,Gao R,Wang Q,Chen H,Zhang S,Mao X,Leblanc M,Behensky A,Zhang Z,Gan L,Yu H,Zhu T,Liu J
Thu Oct 01 00:00:00 EDT 2020
Seryl tRNA synthetase cooperates with POT1 to regulate telomere length and cellular senescence.
Signal transduction and targeted therapy
Li Y,Li X,Cao M,Jiang Y,Yan J,Liu Z,Yang R,Chen X,Sun P,Xiang R,Wang L,Shi Y
Fri Jan 29 00:00:00 EST 2021
Targeting of LRRC59 to the Endoplasmic Reticulum and the Inner Nuclear Membrane.
International journal of molecular sciences
Blenski M,Kehlenbach RH
Tue Jan 15 00:00:00 EST 2019
ASPP2 inhibits tumor growth by repressing the mevalonate pathway in hepatocellular carcinoma.
Cell death & disease
Liang B,Chen R,Song S,Wang H,Sun G,Yang H,Jing W,Zhou X,Fu Z,Huang G,Zhao J
Mon Nov 04 00:00:00 EST 2019
Dichloroacetate Overcomes Oxaliplatin Chemoresistance in Colorectal Cancer through the miR-543/PTEN/Akt/mTOR Pathway.
Journal of Cancer
Liang Y,Zhu D,Zhu L,Hou Y,Hou L,Huang X,Li L,Wang Y,Li L,Zou H,Wu T,Yao M,Wang J,Meng X
Thu Oct 01 00:00:00 EDT 2020
The selective HDAC6 inhibitor Nexturastat A induces apoptosis, overcomes drug resistance and inhibits tumor growth in multiple myeloma.
Bioscience reports
Sun X,Xie Y,Sun X,Yao Y,Li H,Li Z,Yao R,Xu K
Fri Mar 29 00:00:00 EDT 2019
E3 ligase Fbw7 participates in oxidative stress‑induced myocardial cell injury via interacting with Mcl‑1.
Molecular medicine reports
Li X,Zhang N,Zhang Y,Jia P,Guo Y,Tian Y,You S,Wu S,Sun Y
Thu Aug 01 00:00:00 EDT 2019
PTK2 promotes cancer stem cell traits in hepatocellular carcinoma by activating Wnt/β-catenin signaling.
Cancer letters
Fan Z,Duan J,Wang L,Xiao S,Li L,Yan X,Yao W,Wu L,Zhang S,Zhang Y,Li Y,Zhu X,Hu Y,Zhang D,Jiao S,Xu X
Tue May 28 00:00:00 EDT 2019
Resveratrol induced premature senescence and inhibited epithelial-mesenchymal transition of cancer cells via induction of tumor suppressor Rad9.
PloS one
Chen KY,Chen CC,Chang YC,Chang MC
Mon Mar 16 00:00:00 EDT 2020
GADD45a Mediated Cell Cycle Inhibition Is Regulated By P53 In Bladder Cancer.
OncoTargets and therapy
Han N,Yuan F,Xian P,Liu N,Liu J,Zhang H,Zhang H,Yao K,Yuan G
Sun Apr 10 00:00:00 EDT 2022
Histone Demethylase UTX is an Essential Factor for Zygotic Genome Activation and Regulates Zscan4 Expression in Mouse Embryos.
International journal of biological sciences
Bai L,Yang L,Zhao C,Song L,Liu X,Bai C,Su G,Wei Z,Li G
Thu May 07 00:00:00 EDT 2020
Kallikrein 12 Regulates Innate Resistance of Murine Macrophages against Mycobacterium bovis Infection by Modulating Autophagy and Apoptosis.
Cells
Sabir N,Hussain T,Liao Y,Wang J,Song Y,Shahid M,Cheng G,Mangi MH,Yao J,Yang L,Zhao D,Zhou X
Sun May 05 00:00:00 EDT 2019
Impaired Mitochondrial Fusion and Oxidative Phosphorylation Triggered by High Glucose Is Mediated by Tom22 in Endothelial Cells.
Oxidative medicine and cellular longevity
Zeng Y,Pan Q,Wang X,Li D,Lin Y,Man F,Xiao F,Guo L
Fri Dec 13 00:00:00 EST 2019
Inhibition of dendritic spine extension through acrolein conjugation with α-, β-tubulin proteins.
The international journal of biochemistry & cell biology
Uemura T,Suzuki T,Ko K,Watanabe K,Dohmae N,Sakamoto A,Terui Y,Toida T,Kashiwagi K,Igarashi K
Thu Aug 01 00:00:00 EDT 2019
Osteoglycin (OGN) Inhibits Cell Proliferation and Invasiveness in Breast Cancer via PI3K/Akt/mTOR Signaling Pathway.
OncoTargets and therapy
Xu T,Zhang R,Dong M,Zhang Z,Li H,Zhan C,Li X
Mon Apr 11 00:00:00 EDT 2022
HOXA9 is a novel myopia risk gene.
BMC ophthalmology
Liang CL,Hsu PY,Ngo CS,Seow WJ,Karnani N,Pan H,Saw SM,Juo SH
Wed Jan 23 00:00:00 EST 2019
Involvement of STAT3 in the synergistic induction of 11β-HSD1 by SAA1 and cortisol in human amnion fibroblasts.
American journal of reproductive immunology (New York, N.Y. : 1989)
Lu Y,Zhou Q,Lu JW,Wang WS,Sun K
Thu Aug 01 00:00:00 EDT 2019
Crosstalk between Cdk5/p35 and ERK1/2 signalling mediates spinal astrocyte activity via the PPARγ pathway in a rat model of chronic constriction injury.
Journal of neurochemistry
Zhong Y,Chen J,Chen J,Chen Y,Li L,Xie Y
Tue Oct 01 00:00:00 EDT 2019
Polypeptide N-acetylgalactosaminyltransferase 18 non-catalytically regulates the ER homeostasis and O-glycosylation.
Biochimica et biophysica acta. General subjects
Shan A,Lu J,Xu Z,Li X,Xu Y,Li W,Liu F,Yang F,Sato T,Narimatsu H,Zhang Y
Wed May 01 00:00:00 EDT 2019
Ubiquinol-cytochrome C reductase core protein II promotes tumorigenesis by facilitating p53 degradation.
EBioMedicine
Han Y,Wu P,Wang Z,Zhang Z,Sun S,Liu J,Gong S,Gao P,Iwakuma T,Molina-Vila MA,Chen BP,Zhang Y,Ji T,Mo Q,Chen P,Hu J,Wang S,Zhou J,Lu H,Gao Q
Fri Feb 01 00:00:00 EST 2019
In situ repurposing of dendritic cells with CRISPR/Cas9-based nanomedicine to induce transplant tolerance.
Biomaterials
Zhang Y,Shen S,Zhao G,Xu CF,Zhang HB,Luo YL,Cao ZT,Shi J,Zhao ZB,Lian ZX,Wang J
Tue Oct 01 00:00:00 EDT 2019
Antisense oligonucleotide treatment ameliorates IFN-γ-induced proteinuria in APOL1-transgenic mice.
JCI insight
Aghajan M,Booten SL,Althage M,Hart CE,Ericsson A,Maxvall I,Ochaba J,Menschik-Lundin A,Hartleib J,Kuntz S,Gattis D,Ahlström C,Watt AT,Engelhardt JA,Monia BP,Magnone MC,Guo S
Thu Jun 20 00:00:00 EDT 2019
Palmitoylome profiling indicates that androgens regulate the palmitoylation of α‑tubulin in prostate cancer‑derived LNCaP cells and supernatants.
Oncology reports
Li W,Zhang J,Zou L,Cui J,Su F,Jin J,Xiao F,Liu M,Zhao G
Sun Dec 01 00:00:00 EST 2019
Validation of Phosphodiesterase-10 as a Novel Target for Pulmonary Arterial Hypertension via Highly Selective and Subnanomolar Inhibitors.
Journal of medicinal chemistry
Huang YY,Yu YF,Zhang C,Chen Y,Zhou Q,Li Z,Zhou S,Li Z,Guo L,Wu D,Wu Y,Luo HB
Thu Apr 11 00:00:00 EDT 2019
Total flavonoids from sea buckthorn ameliorates lipopolysaccharide/cigarette smoke-induced airway inflammation.
Phytotherapy research : PTR
Ren QC,Li XH,Li QY,Yang HL,Wang HL,Zhang H,Zhao L,Jiang-Yong SL,Meng XL,Zhang Y,Shen XF
Thu Aug 01 00:00:00 EDT 2019
Functional role of GKAP1 in the regulation of male germ cell spontaneous apoptosis and sperm number.
Molecular reproduction and development
Wang H,Wang G,Dai Y,Li Z,Zhu Y,Sun F
Sun Sep 01 00:00:00 EDT 2019
Extracellular RNAs-TLR3 signaling contributes to cognitive decline in a mouse model of postoperative cognitive dysfunction.
Brain, behavior, and immunity
Chen C,Gao R,Li M,Wang Q,Chen H,Zhang S,Mao X,Behensky A,Zhang Z,Gan L,Li T,Liao R,Li Q,Yu H,Yang J,Zhu T,Liu J
Thu Aug 01 00:00:00 EDT 2019
Dorsomorphin induces cancer cell apoptosis and sensitizes cancer cells to HSP90 and proteasome inhibitors by reducing nuclear heat shock factor 1 levels.
Cancer biology & medicine
Li N,Wang T,Li Z,Ye X,Deng B,Zhuo S,Yao P,Yang M,Mei H,Chen X,Zhu T,Chen S,Wang H,Wang J,Le Y
Wed May 01 00:00:00 EDT 2019
Adenosine Attenuates Aortic Smooth Muscle Cell Calcification through A3 Adenosine Receptor.
The Tohoku journal of experimental medicine
Fujimoto K,Shioi A,Miki Y,Kakutani Y,Morioka T,Shoji T,Emoto M,Inaba M
Sun Dec 01 00:00:00 EST 2019
Targeting Super-Enhancer-Driven Oncogenic Transcription by CDK7 Inhibition in Anaplastic Thyroid Carcinoma.
Thyroid : official journal of the American Thyroid Association
Cao X,Dang L,Zheng X,Lu Y,Lu Y,Ji R,Zhang T,Ruan X,Zhi J,Hou X,Yi X,Li MJ,Gu T,Gao M,Zhang L,Chen Y
Sat Jun 01 00:00:00 EDT 2019
CXCR7 regulates epileptic seizures by controlling the synaptic activity of hippocampal granule cells.
Cell death & disease
Xu T,Yu X,Deng J,Ou S,Liu X,Wang T,Liu Y,Yang J,Tan C,Yuan J,Chen Y
Thu Oct 31 00:00:00 EDT 2019
Calcium channel blockers reduce severe fever with thrombocytopenia syndrome virus (SFTSV) related fatality.
Cell research
Li H,Zhang LK,Li SF,Zhang SF,Wan WW,Zhang YL,Xin QL,Dai K,Hu YY,Wang ZB,Zhu XT,Fang YJ,Cui N,Zhang PH,Yuan C,Lu QB,Bai JY,Deng F,Xiao GF,Liu W,Peng K
Sun Sep 01 00:00:00 EDT 2019
PAQR3 regulates phosphorylation of FoxO1 in insulin-resistant HepG2 cells via NF-κB signaling pathway.
Experimental cell research
Chen L,Sun X,Xiao H,Xu F,Yang Y,Lin Z,Chen Z,Quan S,Huang H
Thu Aug 15 00:00:00 EDT 2019
Initial human T-cell leukemia virus type 1 infection of the salivary gland epithelial cells requires a biofilm-like structure.
Virus research
Nakamura H,Shimizu T,Takatani A,Suematsu T,Nakamura T,Kawakami A
Thu Aug 01 00:00:00 EDT 2019
Estrogen negatively regulates the renal epithelial sodium channel (ENaC) by promoting Derlin-1 expression and AMPK activation.
Experimental & molecular medicine
Zhang X,Ge Y,Bukhari AA,Zhu Q,Shen Y,Li M,Sun H,Su D,Liang X
Tue May 21 00:00:00 EDT 2019
Identification of serum metabolites associating with chronic kidney disease progression and anti-fibrotic effect of 5-methoxytryptophan.
Nature communications
Chen DQ,Cao G,Chen H,Argyopoulos CP,Yu H,Su W,Chen L,Samuels DC,Zhuang S,Bayliss GP,Zhao S,Yu XY,Vaziri ND,Wang M,Liu D,Mao JR,Ma SX,Zhao J,Zhang Y,Shang YQ,Kang H,Ye F,Cheng XH,Li XR,Zhang L,Meng MX,Guo Y,Zhao YY
Mon Apr 01 00:00:00 EDT 2019
Major vault protein suppresses obesity and atherosclerosis through inhibiting IKK-NF-κB signaling mediated inflammation.
Nature communications
Ben J,Jiang B,Wang D,Liu Q,Zhang Y,Qi Y,Tong X,Chen L,Liu X,Zhang Y,Zhu X,Li X,Zhang H,Bai H,Yang Q,Ma J,Wiemer EAC,Xu Y,Chen Q
Wed Apr 17 00:00:00 EDT 2019
An alternative membrane topology permits lipid droplet localization of peroxisomal fatty acyl-CoA reductase 1.
Journal of cell science
Exner T,Romero-Brey I,Yifrach E,Rivera-Monroy J,Schrul B,Zouboulis CC,Stremmel W,Honsho M,Bartenschlager R,Zalckvar E,Poppelreuther M,Füllekrug J
Mon Mar 18 00:00:00 EDT 2019
SIRT6/HIF-1α axis promotes papillary thyroid cancer progression by inducing epithelial-mesenchymal transition.
Cancer cell international
Yang Z,Yu W,Huang R,Ye M,Min Z
Wed Aug 23 00:00:00 EDT 2023
A technique for delineating the unfolding requirements for substrate entry into retrotranslocons during endoplasmic reticulum-associated degradation.
The Journal of biological chemistry
Shi J,Hu X,Guo Y,Wang L,Ji J,Li J,Zhang ZR
Fri Dec 27 00:00:00 EST 2019
Endoplasmic reticulum stress, a novel significant mechanism responsible for DEHP-induced increased distance between seminiferous tubule of mouse testis.
Journal of cellular physiology
Pan J,Yao Y,Guo X,Kong F,Zhou J,Meng X
Fri Nov 01 00:00:00 EDT 2019
Targeting JUN, CEBPB, and HDAC3: A Novel Strategy to Overcome Drug Resistance in Hypoxic Glioblastoma.
Frontiers in oncology
Gao Y,Liu B,Feng L,Sun B,He S,Yang Y,Wu G,E G,Liu C,Gao Y,Zhang E,Zhu B
Thu Oct 05 00:00:00 EDT 2023
Small GTPase Arf6 regulates diabetes-induced cholesterol accumulation in podocytes.
Journal of cellular physiology
Hu J,Yang Q,Chen Z,Liang W,Feng J,Ding G
Sun Dec 01 00:00:00 EST 2019
Combined melatonin and poricoic acid A inhibits renal fibrosis through modulating the interaction of Smad3 and β-catenin pathway in AKI-to-CKD continuum.
Therapeutic advances in chronic disease
Chen DQ,Cao G,Zhao H,Chen L,Yang T,Wang M,Vaziri ND,Guo Y,Zhao YY
Sun Apr 10 00:00:00 EDT 2022
Deficiency of ZnT8 Promotes Adiposity and Metabolic Dysfunction by Increasing Peripheral Serotonin Production.
Diabetes
Mao Z,Lin H,Su W,Li J,Zhou M,Li Z,Zhou B,Yang Q,Zhou M,Pan K,He J,Zhang W
Sat Jun 01 00:00:00 EDT 2019
NEDD4 protects vascular endothelial cells against Angiotensin II-induced cell death via enhancement of XPO1-mediated nuclear export.
Experimental cell research
Xu J,Sheng Z,Li F,Wang S,Yuan Y,Wang M,Yu Z
Tue Oct 01 00:00:00 EDT 2019
Microtubule-associated protein 4 phosphorylation regulates epidermal keratinocyte migration and proliferation.
International journal of biological sciences
Zhang J,Li L,Zhang Q,Wang W,Zhang D,Jia J,Lv Y,Yuan H,Song H,Xiang F,Hu J,Huang Y
Mon Apr 13 00:00:00 EDT 2020
Interaction of hnRNP K with MAP 1B-LC1 promotes TGF-β1-mediated epithelial to mesenchymal transition in lung cancer cells.
BMC cancer
Li L,Yan S,Zhang H,Zhang M,Huang G,Chen M
Fri Sep 06 00:00:00 EDT 2019
Isosteviol Sodium Protects Neural Cells Against Hypoxia-Induced Apoptosis Through Inhibiting MAPK and NF-κB Pathways.
Journal of stroke and cerebrovascular diseases : the official journal of National Stroke Association
Zhong KL,Lu MY,Liu F,Mei Y,Zhang XJ,Zhang H,Zan J,Sun XO,Tan W
Tue Jan 01 00:00:00 EST 2019
Dendrobium officinale polysaccharides regulate age-related lineage commitment between osteogenic and adipogenic differentiation.
Cell proliferation
Peng H,Yang M,Guo Q,Su T,Xiao Y,Xia ZY
Mon Jul 01 00:00:00 EDT 2019
Deficiency of PTEN leads to aberrant chromosome segregation through downregulation of MAD2.
Molecular medicine reports
Sun Z,Lu J,Wu M,Li M,Bai L,Shi Z,Hao L,Wu Y
Fri Nov 01 00:00:00 EDT 2019
miR-125b-5p targeting TRAF6 relieves skeletal muscle atrophy induced by fasting or denervation.
Annals of translational medicine
Qiu J,Zhu J,Zhang R,Liang W,Ma W,Zhang Q,Huang Z,Ding F,Sun H
Sun Sep 01 00:00:00 EDT 2019
Esrrb plays important roles in maintaining self-renewal of trophoblast stem cells (TSCs) and reprogramming somatic cells to induced TSCs.
Journal of molecular cell biology
Gao H,Gao R,Zhang L,Xiu W,Zang R,Wang H,Zhang Y,Chen J,Gao Y,Gao S
Sat Jun 01 00:00:00 EDT 2019
SHIP1 inhibits cell growth, migration, and invasion in non‑small cell lung cancer through the PI3K/AKT pathway.
Oncology reports
Fu Q,Huang Y,Ge C,Li Z,Tian H,Li Q,Li H,Li R,Tao X,Xue Y,Wang Y,Yang G,Fang W,Song X
Mon Apr 01 00:00:00 EDT 2019
Cardiac proteomics reveals the potential mechanism of microtubule associated protein 4 phosphorylation-induced mitochondrial dysfunction.
Burns & trauma
Li L,Zhang J,Zhang Q,Huang Y,Hu J
Fri Apr 08 00:00:00 EDT 2022
Cancer‑associated adipocytes exhibit distinct phenotypes and facilitate tumor progression in pancreatic cancer.
Oncology reports
Cai Z,Liang Y,Xing C,Wang H,Hu P,Li J,Huang H,Wang W,Jiang C
Sun Dec 01 00:00:00 EST 2019
UNBS5162 inhibits SKOV3 ovarian cancer cell proliferation by regulating the PI3K/AKT signalling pathway.
Oncology letters
Wang Q,Shi W
Fri Mar 01 00:00:00 EST 2019
Cancer-associated fibroblasts promote colorectal cancer progression by secreting CLEC3B.
Cancer biology & therapy
Zhu HF,Zhang XH,Gu CS,Zhong Y,Long T,Ma YD,Hu ZY,Li ZG,Wang XY
Wed Jul 29 00:00:00 EDT 2020
Targeting CDK12-mediated transcription regulation in anaplastic thyroid carcinoma.
Biochemical and biophysical research communications
Geng M,Yang Y,Cao X,Dang L,Zhang T,Zhang L
Tue Dec 10 00:00:00 EST 2019
Identification of Aloperine as an anti-apoptotic Bcl2 protein inhibitor in glioma cells.
PeerJ
Xu Z,Wang X,Chen X,Zeng S,Qian L,Wei J,Gong Z,Yan Y
Sun Apr 10 00:00:00 EDT 2022
Mitochondrial NOS1 suppresses apoptosis in colon cancer cells through increasing SIRT3 activity.
Biochemical and biophysical research communications
Wang Q,Ye S,Chen X,Xu P,Li K,Zeng S,Huang M,Gao W,Chen J,Zhang Q,Zhong Z,Liu Q
Tue Aug 06 00:00:00 EDT 2019
Bioinformatics Analysis Identifies Protein Tyrosine Kinase 7 (PTK7) as a Potential Prognostic and Therapeutic Biomarker in Stages I to IV Hepatocellular Carcinoma.
Medical science monitor : international medical journal of experimental and clinical research
Zou RC,Liang Y,Li LL,Tang JZ,Yang YP,Geng YC,He J,Luo LY,Li WX,Sun ZW,Yuan HL
Fri Nov 15 00:00:00 EST 2019
Leonurine alleviates LPS-induced myocarditis through suppressing the NF-кB signaling pathway.
Toxicology
Wang R,Li D,Ouyang J,Tian X,Zhao Y,Peng X,Li S,Yu G,Yang J
Sat Jun 15 00:00:00 EDT 2019
Activity-based E3 ligase profiling uncovers an E3 ligase with esterification activity.
Nature
Pao KC,Wood NT,Knebel A,Rafie K,Stanley M,Mabbitt PD,Sundaramoorthy R,Hofmann K,van Aalten DMF,Virdee S
Sun Apr 01 00:00:00 EDT 2018
Activity-based E3 ligase profiling uncovers an E3 ligase with esterification activity.
Nature
Pao KC,Wood NT,Knebel A,Rafie K,Stanley M,Mabbitt PD,Sundaramoorthy R,Hofmann K,van Aalten DMF,Virdee S
Sun Apr 01 00:00:00 EDT 2018
Transcriptional Regulation of the Warburg Effect in Cancer by SIX1.
Cancer cell
Li L,Liang Y,Kang L,Liu Y,Gao S,Chen S,Li Y,You W,Dong Q,Hong T,Yan Z,Jin S,Wang T,Zhao W,Mai H,Huang J,Han X,Ji Q,Song Q,Yang C,Zhao S,Xu X,Ye Q
Mon Mar 12 00:00:00 EDT 2018
Transcriptional Regulation of the Warburg Effect in Cancer by SIX1.
Cancer cell
Li L,Liang Y,Kang L,Liu Y,Gao S,Chen S,Li Y,You W,Dong Q,Hong T,Yan Z,Jin S,Wang T,Zhao W,Mai H,Huang J,Han X,Ji Q,Song Q,Yang C,Zhao S,Xu X,Ye Q
Mon Mar 12 00:00:00 EDT 2018
Basolateral CD147 induces hepatocyte polarity loss by E-cadherin ubiquitination and degradation in hepatocellular carcinoma progress.
Hepatology (Baltimore, Md.)
Lu M,Wu J,Hao ZW,Shang YK,Xu J,Nan G,Li X,Chen ZN,Bian H
Sun Jul 01 00:00:00 EDT 2018
Developmental History Provides a Roadmap for the Emergence of Tumor Plasticity.
Developmental cell
Tata PR,Chow RD,Saladi SV,Tata A,Konkimalla A,Bara A,Montoro D,Hariri LP,Shih AR,Mino-Kenudson M,Mou H,Kimura S,Ellisen LW,Rajagopal J
Mon Mar 26 00:00:00 EDT 2018
Developmental History Provides a Roadmap for the Emergence of Tumor Plasticity.
Developmental cell
Tata PR,Chow RD,Saladi SV,Tata A,Konkimalla A,Bara A,Montoro D,Hariri LP,Shih AR,Mino-Kenudson M,Mou H,Kimura S,Ellisen LW,Rajagopal J
Mon Mar 26 00:00:00 EDT 2018
Sofosbuvir inhibits hepatitis A virus replication in vitro assessed by a cell-based fluorescent reporter system.
Antiviral research
Jiang W,Muhammad F,Ma P,Liu X,Long G
Fri Jun 01 00:00:00 EDT 2018
Sofosbuvir inhibits hepatitis A virus replication in vitro assessed by a cell-based fluorescent reporter system.
Antiviral research
Jiang W,Muhammad F,Ma P,Liu X,Long G
Fri Jun 01 00:00:00 EDT 2018
Novel inhibitors of the cellular renin-angiotensin system components, poricoic acids, target Smad3 phosphorylation and Wnt/β-catenin pathway against renal fibrosis.
British journal of pharmacology
Wang M,Chen DQ,Chen L,Cao G,Zhao H,Liu D,Vaziri ND,Guo Y,Zhao YY
Sun Jul 01 00:00:00 EDT 2018
Novel inhibitors of the cellular renin-angiotensin system components, poricoic acids, target Smad3 phosphorylation and Wnt/β-catenin pathway against renal fibrosis.
British journal of pharmacology
Wang M,Chen DQ,Chen L,Cao G,Zhao H,Liu D,Vaziri ND,Guo Y,Zhao YY
Sun Jul 01 00:00:00 EDT 2018
Interferon-alpha enhances the antitumour activity of EGFR-targeted therapies by upregulating RIG-I in head and neck squamous cell carcinoma.
British journal of cancer
Ma H,Jin S,Yang W,Zhou G,Zhao M,Fang S,Zhang Z,Hu J
Tue Feb 20 00:00:00 EST 2018
Interferon-alpha enhances the antitumour activity of EGFR-targeted therapies by upregulating RIG-I in head and neck squamous cell carcinoma.
British journal of cancer
Ma H,Jin S,Yang W,Zhou G,Zhao M,Fang S,Zhang Z,Hu J
Tue Feb 20 00:00:00 EST 2018
β-catenin regulates IRF3-mediated innate immune signalling in colorectal cancer.
Cell proliferation
Ding C,He J,Zhao J,Li J,Chen J,Liao W,Zeng Y,Zhong J,Wei C,Zhang L,Zhou M,Jia Z,Zhang Y,Li H,Zhou Y,Xiao X,Han D,Li C,Zhu Z,Xia Z,Peng J
Mon Oct 01 00:00:00 EDT 2018
β-catenin regulates IRF3-mediated innate immune signalling in colorectal cancer.
Cell proliferation
Ding C,He J,Zhao J,Li J,Chen J,Liao W,Zeng Y,Zhong J,Wei C,Zhang L,Zhou M,Jia Z,Zhang Y,Li H,Zhou Y,Xiao X,Han D,Li C,Zhu Z,Xia Z,Peng J
Mon Oct 01 00:00:00 EDT 2018
Carvedilol attenuates liver fibrosis by suppressing autophagy and promoting apoptosis in hepatic stellate cells.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Meng D,Li Z,Wang G,Ling L,Wu Y,Zhang C
Sat Dec 01 00:00:00 EST 2018
Carvedilol attenuates liver fibrosis by suppressing autophagy and promoting apoptosis in hepatic stellate cells.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Meng D,Li Z,Wang G,Ling L,Wu Y,Zhang C
Sat Dec 01 00:00:00 EST 2018
Resveratrol-Induced Downregulation of NAF-1 Enhances the Sensitivity of Pancreatic Cancer Cells to Gemcitabine via the ROS/Nrf2 Signaling Pathways.
Oxidative medicine and cellular longevity
Cheng L,Yan B,Chen K,Jiang Z,Zhou C,Cao J,Qian W,Li J,Sun L,Ma J,Ma Q,Sha H
Fri Sep 21 00:00:00 EDT 2018
Resveratrol-Induced Downregulation of NAF-1 Enhances the Sensitivity of Pancreatic Cancer Cells to Gemcitabine via the ROS/Nrf2 Signaling Pathways.
Oxidative medicine and cellular longevity
Cheng L,Yan B,Chen K,Jiang Z,Zhou C,Cao J,Qian W,Li J,Sun L,Ma J,Ma Q,Sha H
Fri Sep 21 00:00:00 EDT 2018
Design, synthesis, biological evaluation and cocrystal structures with tubulin of chiral β-lactam bridged combretastatin A-4 analogues as potent antitumor agents.
European journal of medicinal chemistry
Zhou P,Liang Y,Zhang H,Jiang H,Feng K,Xu P,Wang J,Wang X,Ding K,Luo C,Liu M,Wang Y
Sat Jan 20 00:00:00 EST 2018
Design, synthesis, biological evaluation and cocrystal structures with tubulin of chiral β-lactam bridged combretastatin A-4 analogues as potent antitumor agents.
European journal of medicinal chemistry
Zhou P,Liang Y,Zhang H,Jiang H,Feng K,Xu P,Wang J,Wang X,Ding K,Luo C,Liu M,Wang Y
Sat Jan 20 00:00:00 EST 2018
Redox-active trace metal-induced release of high mobility group box 1(HMGB1) and inflammatory cytokines in fibroblast-like synovial cells is Toll-like receptor 4 (TLR4) dependent.
Biochimica et biophysica acta. Molecular basis of disease
Alsousi AA,Igwe OJ
Thu Nov 01 00:00:00 EDT 2018
Human Cytomegalovirus Protein pUL38 Prevents Premature Cell Death by Binding to Ubiquitin-Specific Protease 24 and Regulating Iron Metabolism.
Journal of virology
Sun Y,Bao Q,Xuan B,Xu W,Pan D,Li Q,Qian Z
Sun Jul 01 00:00:00 EDT 2018
Human Cytomegalovirus Protein pUL38 Prevents Premature Cell Death by Binding to Ubiquitin-Specific Protease 24 and Regulating Iron Metabolism.
Journal of virology
Sun Y,Bao Q,Xuan B,Xu W,Pan D,Li Q,Qian Z
Sun Jul 01 00:00:00 EDT 2018
The protective effects of distal ischemic treatment on apoptosis and mitochondrial permeability in the hippocampus after cardiopulmonary resuscitation.
Journal of cellular physiology
Zhou X,Yong L,Huang Y,Zhu S,Song X,Li B,Zhu J,Wang H
Sat Sep 01 00:00:00 EDT 2018
The protective effects of distal ischemic treatment on apoptosis and mitochondrial permeability in the hippocampus after cardiopulmonary resuscitation.
Journal of cellular physiology
Zhou X,Yong L,Huang Y,Zhu S,Song X,Li B,Zhu J,Wang H
Sat Sep 01 00:00:00 EDT 2018
Cisplatin and curcumin co-loaded nano-liposomes for the treatment of hepatocellular carcinoma.
International journal of pharmaceutics
Cheng Y,Zhao P,Wu S,Yang T,Chen Y,Zhang X,He C,Zheng C,Li K,Ma X,Xiang G
Tue Jul 10 00:00:00 EDT 2018
Cisplatin and curcumin co-loaded nano-liposomes for the treatment of hepatocellular carcinoma.
International journal of pharmaceutics
Cheng Y,Zhao P,Wu S,Yang T,Chen Y,Zhang X,He C,Zheng C,Li K,Ma X,Xiang G
Tue Jul 10 00:00:00 EDT 2018
Epigenetic activation of PERP transcription by MKL1 contributes to ROS-induced apoptosis in skeletal muscle cells.
Biochimica et biophysica acta. Gene regulatory mechanisms
Li Z,Chen B,Dong W,Xu W,Song M,Fang M,Guo J,Xu Y
Fri Jul 27 00:00:00 EDT 2018
Epigenetic activation of PERP transcription by MKL1 contributes to ROS-induced apoptosis in skeletal muscle cells.
Biochimica et biophysica acta. Gene regulatory mechanisms
Li Z,Chen B,Dong W,Xu W,Song M,Fang M,Guo J,Xu Y
Fri Jul 27 00:00:00 EDT 2018
Exogenous oxidants activate nuclear factor kappa B through Toll-like receptor 4 stimulation to maintain inflammatory phenotype in macrophage.
Biochemical pharmacology
Zhang Y,Igwe OJ
Mon Jan 01 00:00:00 EST 2018
Exogenous oxidants activate nuclear factor kappa B through Toll-like receptor 4 stimulation to maintain inflammatory phenotype in macrophage.
Biochemical pharmacology
Zhang Y,Igwe OJ
Mon Jan 01 00:00:00 EST 2018
Novel RAS Inhibitors Poricoic Acid ZG and Poricoic Acid ZH Attenuate Renal Fibrosis via a Wnt/β-Catenin Pathway and Targeted Phosphorylation of smad3 Signaling.
Journal of agricultural and food chemistry
Wang M,Chen DQ,Chen L,Liu D,Zhao H,Zhang ZH,Vaziri ND,Guo Y,Zhao YY,Cao G
Wed Feb 28 00:00:00 EST 2018
Novel RAS Inhibitors Poricoic Acid ZG and Poricoic Acid ZH Attenuate Renal Fibrosis via a Wnt/β-Catenin Pathway and Targeted Phosphorylation of smad3 Signaling.
Journal of agricultural and food chemistry
Wang M,Chen DQ,Chen L,Liu D,Zhao H,Zhang ZH,Vaziri ND,Guo Y,Zhao YY,Cao G
Wed Feb 28 00:00:00 EST 2018
Immune Activation of RAW264.7 Macrophages by Low Molecular Weight Fucoidan Extracted from New Zealand Undaria pinnatifida.
Journal of agricultural and food chemistry
Bi D,Yu B,Han Q,Lu J,White WL,Lai Q,Cai N,Luo W,Gu L,Li S,Xu H,Hu Z,Nie S,Xu X
Wed Oct 17 00:00:00 EDT 2018
Immune Activation of RAW264.7 Macrophages by Low Molecular Weight Fucoidan Extracted from New Zealand Undaria pinnatifida.
Journal of agricultural and food chemistry
Bi D,Yu B,Han Q,Lu J,White WL,Lai Q,Cai N,Luo W,Gu L,Li S,Xu H,Hu Z,Nie S,Xu X
Wed Oct 17 00:00:00 EDT 2018
Dynasore-induced potent ubiquitylation of the exon 19 deletion mutant of epidermal growth factor receptor suppresses cell growth and migration in non-small cell lung cancer.
The international journal of biochemistry & cell biology
Wang T,Wang D,Zhang Y,Zhang J,Sun X,Wu Y,Wang S,Zhang Y,Xu L,Kong Q,Gao Y,Wu Y,Liu F,Liu S,Zhang Y,Lei T,Liu H
Sat Dec 01 00:00:00 EST 2018
Dynasore-induced potent ubiquitylation of the exon 19 deletion mutant of epidermal growth factor receptor suppresses cell growth and migration in non-small cell lung cancer.
The international journal of biochemistry & cell biology
Wang T,Wang D,Zhang Y,Zhang J,Sun X,Wu Y,Wang S,Zhang Y,Xu L,Kong Q,Gao Y,Wu Y,Liu F,Liu S,Zhang Y,Lei T,Liu H
Sat Dec 01 00:00:00 EST 2018
Cell division cycle 7 is a potential therapeutic target in oral squamous cell carcinoma and is regulated by E2F1.
Journal of molecular medicine (Berlin, Germany)
Jin S,Ma H,Yang W,Ju H,Wang L,Zhang Z
Fri Jun 01 00:00:00 EDT 2018
Cell division cycle 7 is a potential therapeutic target in oral squamous cell carcinoma and is regulated by E2F1.
Journal of molecular medicine (Berlin, Germany)
Jin S,Ma H,Yang W,Ju H,Wang L,Zhang Z
Fri Jun 01 00:00:00 EDT 2018
Ubiquitin-specific protease 11 serves as a marker of poor prognosis and promotes metastasis in hepatocellular carcinoma.
Laboratory investigation; a journal of technical methods and pathology
Zhang S,Xie C,Li H,Zhang K,Li J,Wang X,Yin Z
Sun Jul 01 00:00:00 EDT 2018
Ubiquitin-specific protease 11 serves as a marker of poor prognosis and promotes metastasis in hepatocellular carcinoma.
Laboratory investigation; a journal of technical methods and pathology
Zhang S,Xie C,Li H,Zhang K,Li J,Wang X,Yin Z
Sun Jul 01 00:00:00 EDT 2018
An adenosine A1R-A2aR imbalance regulates low glucose/hypoxia-induced microglial activation, thereby contributing to oligodendrocyte damage through NF-κB and CREB phosphorylation.
International journal of molecular medicine
Huang W,Bai S,Zuo X,Tang W,Chen P,Chen X,Wang G,Wang H,Xie P
Fri Jun 01 00:00:00 EDT 2018
Low-density lipoprotein receptor-related protein 6 regulates alternative pre-mRNA splicing.
Journal of cellular and molecular medicine
Yuan T,Wang S,Hu C,Wu Y,Liang D,Li L,Liu Y,Li J,Chen YH
Mon Oct 01 00:00:00 EDT 2018
Low-density lipoprotein receptor-related protein 6 regulates alternative pre-mRNA splicing.
Journal of cellular and molecular medicine
Yuan T,Wang S,Hu C,Wu Y,Liang D,Li L,Liu Y,Li J,Chen YH
Mon Oct 01 00:00:00 EDT 2018
The oncogenic potentials and diagnostic significance of long non-coding RNA LINC00310 in breast cancer.
Journal of cellular and molecular medicine
Li J,Peng W,Du L,Yang Q,Wang C,Mo YY
Sat Sep 01 00:00:00 EDT 2018
The oncogenic potentials and diagnostic significance of long non-coding RNA LINC00310 in breast cancer.
Journal of cellular and molecular medicine
Li J,Peng W,Du L,Yang Q,Wang C,Mo YY
Sat Sep 01 00:00:00 EDT 2018
Selenium-enriched yeast inhibited β-amyloid production and modulated autophagy in a triple transgenic mouse model of Alzheimer's disease.
Metallomics : integrated biometal science
Song GL,Chen C,Wu QY,Zhang ZH,Zheng R,Chen Y,Jia SZ,Ni JZ
Wed Aug 15 00:00:00 EDT 2018
Selenium-enriched yeast inhibited β-amyloid production and modulated autophagy in a triple transgenic mouse model of Alzheimer's disease.
Metallomics : integrated biometal science
Song GL,Chen C,Wu QY,Zhang ZH,Zheng R,Chen Y,Jia SZ,Ni JZ
Wed Aug 15 00:00:00 EDT 2018
XRCC1 rs1799782 (C194T) polymorphism correlated with tumor metastasis and molecular subtypes in breast cancer.
OncoTargets and therapy
Li Q,Ma R,Zhang M
Wed Oct 04 00:00:00 EDT 2023
XRCC1 rs1799782 (C194T) polymorphism correlated with tumor metastasis and molecular subtypes in breast cancer.
OncoTargets and therapy
Li Q,Ma R,Zhang M
Wed Oct 04 00:00:00 EDT 2023
Targeting LC3 and Beclin-1 autophagy genes suppresses proliferation, survival, migration and invasion by inhibition of Cyclin-D1 and uPAR/Integrin β1/ Src signaling in triple negative breast cancer cells.
Journal of cancer research and clinical oncology
Hamurcu Z,Delibaşı N,Geçene S,Şener EF,Dönmez-Altuntaş H,Özkul Y,Canatan H,Ozpolat B
Thu Mar 01 00:00:00 EST 2018
Targeting LC3 and Beclin-1 autophagy genes suppresses proliferation, survival, migration and invasion by inhibition of Cyclin-D1 and uPAR/Integrin β1/ Src signaling in triple negative breast cancer cells.
Journal of cancer research and clinical oncology
Hamurcu Z,Delibaşı N,Geçene S,Şener EF,Dönmez-Altuntaş H,Özkul Y,Canatan H,Ozpolat B
Thu Mar 01 00:00:00 EST 2018
Nucleoporin35 is a novel microtubule associated protein functioning in oocyte meiotic spindle architecture.
Experimental cell research
Chen F,Jiao XF,Zhang JY,Wu D,Ding ZM,Wang YS,Miao YL,Huo LJ
Mon Oct 15 00:00:00 EDT 2018
Nucleoporin35 is a novel microtubule associated protein functioning in oocyte meiotic spindle architecture.
Experimental cell research
Chen F,Jiao XF,Zhang JY,Wu D,Ding ZM,Wang YS,Miao YL,Huo LJ
Mon Oct 15 00:00:00 EDT 2018
Oocyte-specific deletion of Gsα induces oxidative stress and deteriorates oocyte quality in mice.
Experimental cell research
Xie Y,Wu B,Jin Y,Zhang A,Sun X,Zhang X,Gao X,Dong R,Li H,Gao J
Sat Sep 15 00:00:00 EDT 2018
Oocyte-specific deletion of Gsα induces oxidative stress and deteriorates oocyte quality in mice.
Experimental cell research
Xie Y,Wu B,Jin Y,Zhang A,Sun X,Zhang X,Gao X,Dong R,Li H,Gao J
Sat Sep 15 00:00:00 EDT 2018
TUSC3 accelerates cancer growth and induces epithelial-mesenchymal transition by upregulating claudin-1 in non-small-cell lung cancer cells.
Experimental cell research
Feng S,Zhai J,Lu D,Lin J,Dong X,Liu X,Wu H,Roden AC,Brandi G,Tavolari S,Bille A,Cai K
Sat Dec 15 00:00:00 EST 2018
TUSC3 accelerates cancer growth and induces epithelial-mesenchymal transition by upregulating claudin-1 in non-small-cell lung cancer cells.
Experimental cell research
Feng S,Zhai J,Lu D,Lin J,Dong X,Liu X,Wu H,Roden AC,Brandi G,Tavolari S,Bille A,Cai K
Sat Dec 15 00:00:00 EST 2018
Inhibitor of apoptosis protein‑like protein‑2: A novel growth accelerator for breast cancer cells.
Oncology reports
Zhu L,Zhou W,Zhu X,Xiang S,Wang S,Peng Y,Lu B,Tang P,Chen Q,Wu M,Peng X,Chen Z,Sun Z,Yang K,Xiang M,Yu D
Mon Oct 01 00:00:00 EDT 2018
Inhibitor of apoptosis protein‑like protein‑2: A novel growth accelerator for breast cancer cells.
Oncology reports
Zhu L,Zhou W,Zhu X,Xiang S,Wang S,Peng Y,Lu B,Tang P,Chen Q,Wu M,Peng X,Chen Z,Sun Z,Yang K,Xiang M,Yu D
Mon Oct 01 00:00:00 EDT 2018
HSF4 transcriptional regulates HMOX-1 expression in HLECs.
Gene
Liao S,Qu Z,Li L,Zhou B,Gao M,Huang M,Li D
Sun May 20 00:00:00 EDT 2018
HSF4 transcriptional regulates HMOX-1 expression in HLECs.
Gene
Liao S,Qu Z,Li L,Zhou B,Gao M,Huang M,Li D
Sun May 20 00:00:00 EDT 2018
Liver-enriched activator protein 1 as an isoform of CCAAT/enhancer-binding protein beta suppresses stem cell features of hepatocellular carcinoma.
Cancer management and research
Yang LH,Wang Y,Qiao S,Wang MJ,Chen F,Zi XY,Li JX,Zhang HB,Yu B,Hu YP
Fri Apr 08 00:00:00 EDT 2022
Liver-enriched activator protein 1 as an isoform of CCAAT/enhancer-binding protein beta suppresses stem cell features of hepatocellular carcinoma.
Cancer management and research
Yang LH,Wang Y,Qiao S,Wang MJ,Chen F,Zi XY,Li JX,Zhang HB,Yu B,Hu YP
Fri Apr 08 00:00:00 EDT 2022
High expression of NPRL2 is linked to poor prognosis in patients with prostate cancer.
Human pathology
Chen Z,Luo S,Chen Y,Xie X,Du Z,Jiang L
Fri Jun 01 00:00:00 EDT 2018
High expression of NPRL2 is linked to poor prognosis in patients with prostate cancer.
Human pathology
Chen Z,Luo S,Chen Y,Xie X,Du Z,Jiang L
Fri Jun 01 00:00:00 EDT 2018
Administration of ubiquitin-activating enzyme UBA1 inhibitor PYR-41 attenuates angiotensin II-induced cardiac remodeling in mice.
Biochemical and biophysical research communications
Shu Q,Lai S,Wang XM,Zhang YL,Yang XL,Bi HL,Li HH
Sat Oct 20 00:00:00 EDT 2018
Administration of ubiquitin-activating enzyme UBA1 inhibitor PYR-41 attenuates angiotensin II-induced cardiac remodeling in mice.
Biochemical and biophysical research communications
Shu Q,Lai S,Wang XM,Zhang YL,Yang XL,Bi HL,Li HH
Sat Oct 20 00:00:00 EDT 2018
ERα is a negative regulator of PD-L1 gene transcription in breast cancer.
Biochemical and biophysical research communications
Liu L,Shen Y,Zhu X,Lv R,Li S,Zhang Z,Shi YG,Tan L
Sat Oct 20 00:00:00 EDT 2018
ERα is a negative regulator of PD-L1 gene transcription in breast cancer.
Biochemical and biophysical research communications
Liu L,Shen Y,Zhu X,Lv R,Li S,Zhang Z,Shi YG,Tan L
Sat Oct 20 00:00:00 EDT 2018
XPO1-mediated nuclear export of RNF146 protects from angiotensin II-induced endothelial cellular injury.
Biochemical and biophysical research communications
Sheng Z,Xu Y,Wang S,Yuan Y,Huang T,Lu P
Mon Sep 10 00:00:00 EDT 2018
XPO1-mediated nuclear export of RNF146 protects from angiotensin II-induced endothelial cellular injury.
Biochemical and biophysical research communications
Sheng Z,Xu Y,Wang S,Yuan Y,Huang T,Lu P
Mon Sep 10 00:00:00 EDT 2018
Chronic inhalation of e-cigarette vapor containing nicotine disrupts airway barrier function and induces systemic inflammation and multiorgan fibrosis in mice.
American journal of physiology. Regulatory, integrative and comparative physiology
Crotty Alexander LE,Drummond CA,Hepokoski M,Mathew D,Moshensky A,Willeford A,Das S,Singh P,Yong Z,Lee JH,Vega K,Du A,Shin J,Javier C,Tian J,Brown JH,Breen EC
Fri Jun 01 00:00:00 EDT 2018
Chronic inhalation of e-cigarette vapor containing nicotine disrupts airway barrier function and induces systemic inflammation and multiorgan fibrosis in mice.
American journal of physiology. Regulatory, integrative and comparative physiology
Crotty Alexander LE,Drummond CA,Hepokoski M,Mathew D,Moshensky A,Willeford A,Das S,Singh P,Yong Z,Lee JH,Vega K,Du A,Shin J,Javier C,Tian J,Brown JH,Breen EC
Fri Jun 01 00:00:00 EDT 2018
Suppressed epithelial-mesenchymal transition and cancer stem cell properties mediate the anti-cancer effects of ethyl pyruvate via regulation of the AKT/nuclear factor-κB pathway in prostate cancer cells.
Oncology letters
Huang B,Lv DJ,Wang C,Shu FP,Gong ZC,Xie T,Yu YZ,Song XL,Xie JJ,Li S,Liu YM,Qi H,Zhao SC
Wed Aug 01 00:00:00 EDT 2018
Suppressed epithelial-mesenchymal transition and cancer stem cell properties mediate the anti-cancer effects of ethyl pyruvate via regulation of the AKT/nuclear factor-κB pathway in prostate cancer cells.
Oncology letters
Huang B,Lv DJ,Wang C,Shu FP,Gong ZC,Xie T,Yu YZ,Song XL,Xie JJ,Li S,Liu YM,Qi H,Zhao SC
Wed Aug 01 00:00:00 EDT 2018
Hypothalamic DNA methylation in rats with dihydrotestosterone-induced polycystic ovary syndrome: effects of low-frequency electro-acupuncture.
Experimental physiology
Cui P,Ma T,Tamadon A,Han S,Li B,Chen Z,An X,Shao LR,Wang Y,Feng Y
Sat Dec 01 00:00:00 EST 2018
Hypothalamic DNA methylation in rats with dihydrotestosterone-induced polycystic ovary syndrome: effects of low-frequency electro-acupuncture.
Experimental physiology
Cui P,Ma T,Tamadon A,Han S,Li B,Chen Z,An X,Shao LR,Wang Y,Feng Y
Sat Dec 01 00:00:00 EST 2018
Periostin secreted by cancer-associated fibroblasts promotes cancer stemness in head and neck cancer by activating protein tyrosine kinase 7.
Cell death & disease
Yu B,Wu K,Wang X,Zhang J,Wang L,Jiang Y,Zhu X,Chen W,Yan M
Mon Oct 22 00:00:00 EDT 2018
Periostin secreted by cancer-associated fibroblasts promotes cancer stemness in head and neck cancer by activating protein tyrosine kinase 7.
Cell death & disease
Yu B,Wu K,Wang X,Zhang J,Wang L,Jiang Y,Zhu X,Chen W,Yan M
Mon Oct 22 00:00:00 EDT 2018
TGF-β induces corneal endothelial senescence via increase of mitochondrial reactive oxygen species in chronic corneal allograft failure.
Aging
Li Z,Liu T,Ma J,Guo Q,Ma L,Lv Q,Jiang Y,Wei C,Zhang J
Wed Nov 28 00:00:00 EST 2018
TGF-β induces corneal endothelial senescence via increase of mitochondrial reactive oxygen species in chronic corneal allograft failure.
Aging
Li Z,Liu T,Ma J,Guo Q,Ma L,Lv Q,Jiang Y,Wei C,Zhang J
Wed Nov 28 00:00:00 EST 2018
Microtubule associated protein 4 phosphorylation leads to pathological cardiac remodeling in mice.
EBioMedicine
Li L,Zhang Q,Zhang X,Zhang J,Wang X,Ren J,Jia J,Zhang D,Jiang X,Zhang J,Mei H,Chen B,Hu J,Huang Y
Thu Nov 01 00:00:00 EDT 2018
Microtubule associated protein 4 phosphorylation leads to pathological cardiac remodeling in mice.
EBioMedicine
Li L,Zhang Q,Zhang X,Zhang J,Wang X,Ren J,Jia J,Zhang D,Jiang X,Zhang J,Mei H,Chen B,Hu J,Huang Y
Thu Nov 01 00:00:00 EDT 2018
Microtubule associated protein 4 phosphorylation leads to pathological cardiac remodeling in mice.
EBioMedicine
Li L,Zhang Q,Zhang X,Zhang J,Wang X,Ren J,Jia J,Zhang D,Jiang X,Zhang J,Mei H,Chen B,Hu J,Huang Y
Thu Nov 01 00:00:00 EDT 2018
Microtubule associated protein 4 phosphorylation leads to pathological cardiac remodeling in mice.
EBioMedicine
Li L,Zhang Q,Zhang X,Zhang J,Wang X,Ren J,Jia J,Zhang D,Jiang X,Zhang J,Mei H,Chen B,Hu J,Huang Y
Thu Nov 01 00:00:00 EDT 2018
Elucidation of the Molecular-Mechanisms and In Vivo Evaluation of the Anti-inflammatory Effect of Alginate-Derived Seleno-polymannuronate.
Journal of agricultural and food chemistry
Bi D,Lai Q,Cai N,Li T,Zhang Y,Han Q,Peng Y,Xu H,Lu J,Bao W,Liu Q,Xu X
Wed Mar 07 00:00:00 EST 2018
Effects of Thymoquinone on radiation enteritis in mice.
Scientific reports
Hou Q,Liu L,Dong Y,Wu J,Du L,Dong H,Li D
Thu Oct 11 00:00:00 EDT 2018
HuR regulates telomerase activity through TERC methylation.
Nature communications
Tang H,Wang H,Cheng X,Fan X,Yang F,Zhang M,Chen Y,Tian Y,Liu C,Shao D,Jiang B,Dou Y,Cong Y,Xing J,Zhang X,Yi X,Songyang Z,Ma W,Zhao Y,Wang X,Ma J,Gorospe M,Ju Z,Wang W
Thu Jun 07 00:00:00 EDT 2018
HuR regulates telomerase activity through TERC methylation.
Nature communications
Tang H,Wang H,Cheng X,Fan X,Yang F,Zhang M,Chen Y,Tian Y,Liu C,Shao D,Jiang B,Dou Y,Cong Y,Xing J,Zhang X,Yi X,Songyang Z,Ma W,Zhao Y,Wang X,Ma J,Gorospe M,Ju Z,Wang W
Thu Jun 07 00:00:00 EDT 2018
Annexin A5 regulates hepatocarcinoma malignancy via CRKI/II-DOCK180-RAC1 integrin and MEK-ERK pathways.
Cell death & disease
Sun X,Liu S,Wang J,Wei B,Guo C,Chen C,Sun MZ
Fri May 25 00:00:00 EDT 2018
Annexin A5 regulates hepatocarcinoma malignancy via CRKI/II-DOCK180-RAC1 integrin and MEK-ERK pathways.
Cell death & disease
Sun X,Liu S,Wang J,Wei B,Guo C,Chen C,Sun MZ
Fri May 25 00:00:00 EDT 2018
Klf4 Alleviates Lipopolysaccharide-Induced Inflammation by Inducing Expression of MCP-1 Induced Protein 1 to Deubiquitinate TRAF6.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology
Li Z,Jia Y,Han S,Wang X,Han F,Zhang J,Zhang W,Guan H,Hu D
Fri Aug 17 00:00:00 EDT 2018
Klf4 Alleviates Lipopolysaccharide-Induced Inflammation by Inducing Expression of MCP-1 Induced Protein 1 to Deubiquitinate TRAF6.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology
Li Z,Jia Y,Han S,Wang X,Han F,Zhang J,Zhang W,Guan H,Hu D
Fri Aug 17 00:00:00 EDT 2018
Expression profiling identified IL-8 as a regulator of homotypic cell-in-cell formation.
BMB reports
Ruan B,Wang C,Chen A,Liang J,Niu Z,Zheng Y,Fan J,Gao L,Huang H,Wang X,Sun Q
Wed Aug 01 00:00:00 EDT 2018
Expression profiling identified IL-8 as a regulator of homotypic cell-in-cell formation.
BMB reports
Ruan B,Wang C,Chen A,Liang J,Niu Z,Zheng Y,Fan J,Gao L,Huang H,Wang X,Sun Q
Wed Aug 01 00:00:00 EDT 2018
MiR-384 inhibits the proliferation of colorectal cancer by targeting AKT3.
Cancer cell international
Wang YX,Zhu HF,Zhang ZY,Ren F,Hu YH
Mon Mar 21 00:00:00 EDT 2022
MiR-384 inhibits the proliferation of colorectal cancer by targeting AKT3.
Cancer cell international
Wang YX,Zhu HF,Zhang ZY,Ren F,Hu YH
Mon Mar 21 00:00:00 EDT 2022
Activation of α7 nicotinic acetylcholine receptor alleviates Aβ1-42-induced neurotoxicity via downregulation of p38 and JNK MAPK signaling pathways.
Neurochemistry international
Chang KW,Zong HF,Ma KG,Zhai WY,Yang WN,Hu XD,Xu JH,Chen XL,Ji SF,Qian YH
Thu Nov 01 00:00:00 EDT 2018
Ubiquitin-Specific Protease USP6 Regulates the Stability of the c-Jun Protein.
Molecular and cellular biology
Li L,Yang H,He Y,Li T,Feng J,Chen W,Ao L,Shi X,Lin Y,Liu H,Zheng E,Lin Q,Bu J,Zeng Y,Zheng M,Xu Y,Liao Z,Lin J,Lin D
Mon Jan 15 00:00:00 EST 2018
Atorvastatin Attenuates Myocardial Hypertrophy in Spontaneously Hypertensive Rats via the C/EBPβ/PGC-1α/UCP3 Pathway.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology
Chen Y,Chang Y,Zhang N,Guo X,Sun G,Sun Y
Thu Jul 05 00:00:00 EDT 2018
Atorvastatin Attenuates Myocardial Hypertrophy in Spontaneously Hypertensive Rats via the C/EBPβ/PGC-1α/UCP3 Pathway.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology
Chen Y,Chang Y,Zhang N,Guo X,Sun G,Sun Y
Thu Jul 05 00:00:00 EDT 2018
MicroRNA-93 mediates cabergoline-resistance by targeting ATG7 in prolactinoma.
The Journal of endocrinology
Wu Z,Cai L,Lu J,Wang C,Guan J,Chen X,Wu J,Zheng W,Wu Z,Li Q,Su Z
Sat Sep 01 00:00:00 EDT 2018
Comparison of the Effects and Inhibitory Pathways of the Constituents from Gynostemma pentaphyllum against LPS-Induced Inflammatory Response.
Journal of agricultural and food chemistry
Shen CY,Jiang JG,Shi MM,Yang HL,Wei H,Zhu W
Wed Oct 31 00:00:00 EDT 2018
SIRT6-mediated transcriptional suppression of Txnip is critical for pancreatic beta cell function and survival in mice.
Diabetologia
Qin K,Zhang N,Zhang Z,Nipper M,Zhu Z,Leighton J,Xu K,Musi N,Wang P
Sun Apr 01 00:00:00 EDT 2018
The Protective Effect of Astaxanthin on Cognitive Function via Inhibition of Oxidative Stress and Inflammation in the Brains of Chronic T2DM Rats.
Frontiers in pharmacology
Feng Y,Chu A,Luo Q,Wu M,Shi X,Chen Y
Thu Oct 01 00:00:00 EDT 2020
The Protective Effect of Astaxanthin on Cognitive Function via Inhibition of Oxidative Stress and Inflammation in the Brains of Chronic T2DM Rats.
Frontiers in pharmacology
Feng Y,Chu A,Luo Q,Wu M,Shi X,Chen Y
Thu Oct 01 00:00:00 EDT 2020
Taurocholic acid is an active promoting factor, not just a biomarker of progression of liver cirrhosis: evidence from a human metabolomic study and in vitro experiments.
BMC gastroenterology
Liu Z,Zhang Z,Huang M,Sun X,Liu B,Guo Q,Chang Q,Duan Z
Wed Jul 11 00:00:00 EDT 2018
Taurocholic acid is an active promoting factor, not just a biomarker of progression of liver cirrhosis: evidence from a human metabolomic study and in vitro experiments.
BMC gastroenterology
Liu Z,Zhang Z,Huang M,Sun X,Liu B,Guo Q,Chang Q,Duan Z
Wed Jul 11 00:00:00 EDT 2018
Aging-dependent decrease in the numbers of enteric neurons, interstitial cells of Cajal and expression of connexin43 in various regions of gastrointestinal tract.
Aging
Sun T,Li D,Hu S,Huang L,Sun H,Yang S,Wu B,Ji F,Zhou D
Tue Dec 11 00:00:00 EST 2018
Aging-dependent decrease in the numbers of enteric neurons, interstitial cells of Cajal and expression of connexin43 in various regions of gastrointestinal tract.
Aging
Sun T,Li D,Hu S,Huang L,Sun H,Yang S,Wu B,Ji F,Zhou D
Tue Dec 11 00:00:00 EST 2018
Fucoidan Alleviates Acetaminophen-Induced Hepatotoxicity via Oxidative Stress Inhibition and Nrf2 Translocation.
International journal of molecular sciences
Wang YQ,Wei JG,Tu MJ,Gu JG,Zhang W
Fri Dec 14 00:00:00 EST 2018
Fucoidan Alleviates Acetaminophen-Induced Hepatotoxicity via Oxidative Stress Inhibition and Nrf2 Translocation.
International journal of molecular sciences
Wang YQ,Wei JG,Tu MJ,Gu JG,Zhang W
Fri Dec 14 00:00:00 EST 2018
TRIM31 Deficiency Is Associated with Impaired Glucose Metabolism and Disrupted Gut Microbiota in Mice.
Frontiers in physiology
Cheng J,Xue F,Zhang M,Cheng C,Qiao L,Ma J,Sui W,Xu X,Gao C,Hao P,Zhang M,Zhang Y
Wed Sep 30 00:00:00 EDT 2020
TRIM31 Deficiency Is Associated with Impaired Glucose Metabolism and Disrupted Gut Microbiota in Mice.
Frontiers in physiology
Cheng J,Xue F,Zhang M,Cheng C,Qiao L,Ma J,Sui W,Xu X,Gao C,Hao P,Zhang M,Zhang Y
Wed Sep 30 00:00:00 EDT 2020
hnRNPK modulates selective quality-control autophagy by downregulating the expression of HDAC6 in 293 cells.
International journal of oncology
Li Z,Liu X,Ma J,Zhang T,Gao X,Liu L
Thu Nov 01 00:00:00 EDT 2018
Fas Signaling in Dendritic Cells Mediates Th2 Polarization in HDM-Induced Allergic Pulmonary Inflammation.
Frontiers in immunology
Han M,Hu R,Ma J,Zhang B,Chen C,Li H,Yang J,Huang G
Fri Oct 25 00:00:00 EDT 2019
Caspase-3 Inhibition Attenuates the Cytopathic Effects of EV71 Infection.
Frontiers in microbiology
Song F,Yu X,Zhong T,Wang Z,Meng X,Li Z,Zhang S,Huo W,Liu X,Zhang Y,Zhang W,Yu J
Mon Mar 21 00:00:00 EDT 2022
Caspase-3 Inhibition Attenuates the Cytopathic Effects of EV71 Infection.
Frontiers in microbiology
Song F,Yu X,Zhong T,Wang Z,Meng X,Li Z,Zhang S,Huo W,Liu X,Zhang Y,Zhang W,Yu J
Mon Mar 21 00:00:00 EDT 2022
Role of c-Abl-GSK3β Signaling in MPP+-Induced Autophagy-Lysosomal Dysfunction.
Toxicological sciences : an official journal of the Society of Toxicology
Ren Y,Chen J,Wu X,Gui C,Mao K,Zou F,Li W
Sat Sep 01 00:00:00 EDT 2018
Role of c-Abl-GSK3β Signaling in MPP+-Induced Autophagy-Lysosomal Dysfunction.
Toxicological sciences : an official journal of the Society of Toxicology
Ren Y,Chen J,Wu X,Gui C,Mao K,Zou F,Li W
Sat Sep 01 00:00:00 EDT 2018
CD300A promotes tumor progression by PECAM1, ADCY7 and AKT pathway in acute myeloid leukemia.
Oncotarget
Sun X,Huang S,Wang X,Zhang X,Wang X
Fri Jun 08 00:00:00 EDT 2018
CD300A promotes tumor progression by PECAM1, ADCY7 and AKT pathway in acute myeloid leukemia.
Oncotarget
Sun X,Huang S,Wang X,Zhang X,Wang X
Fri Jun 08 00:00:00 EDT 2018
Aberrant hypermethylation of the HOXD10 gene in papillary thyroid cancer with BRAFV600E mutation.
Oncology reports
Cao YM,Gu J,Zhang YS,Wei WJ,Qu N,Wen D,Liao T,Shi RL,Zhang L,Ji QH,Wang Y,Sun GH,Zhao YX,Wang YJ,Yu J,Zhu YX
Mon Jan 01 00:00:00 EST 2018
Suppression of MAPK11 or HIPK3 reduces mutant Huntingtin levels in Huntington's disease models.
Cell research
Yu M,Fu Y,Liang Y,Song H,Yao Y,Wu P,Yao Y,Pan Y,Wen X,Ma L,Hexige S,Ding Y,Luo S,Lu B
Fri Dec 01 00:00:00 EST 2017
Suppression of MAPK11 or HIPK3 reduces mutant Huntingtin levels in Huntington's disease models.
Cell research
Yu M,Fu Y,Liang Y,Song H,Yao Y,Wu P,Yao Y,Pan Y,Wen X,Ma L,Hexige S,Ding Y,Luo S,Lu B
Fri Dec 01 00:00:00 EST 2017
Regulation of m6A Transcripts by the 3'→5' RNA Helicase YTHDC2 Is Essential for a Successful Meiotic Program in the Mammalian Germline.
Molecular cell
Wojtas MN,Pandey RR,Mendel M,Homolka D,Sachidanandam R,Pillai RS
Thu Oct 19 00:00:00 EDT 2017
Regulation of m6A Transcripts by the 3'→5' RNA Helicase YTHDC2 Is Essential for a Successful Meiotic Program in the Mammalian Germline.
Molecular cell
Wojtas MN,Pandey RR,Mendel M,Homolka D,Sachidanandam R,Pillai RS
Thu Oct 19 00:00:00 EDT 2017
Autoregulation of RBM10 and cross-regulation of RBM10/RBM5 via alternative splicing-coupled nonsense-mediated decay.
Nucleic acids research
Sun Y,Bao Y,Han W,Song F,Shen X,Zhao J,Zuo J,Saffen D,Chen W,Wang Z,You X,Wang Y
Mon Aug 21 00:00:00 EDT 2017
Autoregulation of RBM10 and cross-regulation of RBM10/RBM5 via alternative splicing-coupled nonsense-mediated decay.
Nucleic acids research
Sun Y,Bao Y,Han W,Song F,Shen X,Zhao J,Zuo J,Saffen D,Chen W,Wang Z,You X,Wang Y
Mon Aug 21 00:00:00 EDT 2017
Protein synthesis inhibition and GADD34 control IFN-β heterogeneous expression in response to dsRNA.
The EMBO journal
Dalet A,Argüello RJ,Combes A,Spinelli L,Jaeger S,Fallet M,Vu Manh TP,Mendes A,Perego J,Reverendo M,Camosseto V,Dalod M,Weil T,Santos MA,Gatti E,Pierre P
Wed Mar 15 00:00:00 EDT 2017
Protein synthesis inhibition and GADD34 control IFN-β heterogeneous expression in response to dsRNA.
The EMBO journal
Dalet A,Argüello RJ,Combes A,Spinelli L,Jaeger S,Fallet M,Vu Manh TP,Mendes A,Perego J,Reverendo M,Camosseto V,Dalod M,Weil T,Santos MA,Gatti E,Pierre P
Wed Mar 15 00:00:00 EDT 2017
PADI2-Mediated Citrullination Promotes Prostate Cancer Progression.
Cancer research
Wang L,Song G,Zhang X,Feng T,Pan J,Chen W,Yang M,Bai X,Pang Y,Yu J,Han J,Han B
Wed Nov 01 00:00:00 EDT 2017
PADI2-Mediated Citrullination Promotes Prostate Cancer Progression.
Cancer research
Wang L,Song G,Zhang X,Feng T,Pan J,Chen W,Yang M,Bai X,Pang Y,Yu J,Han J,Han B
Wed Nov 01 00:00:00 EDT 2017
Cell-autonomous adiposity through increased cell surface GLUT4 due to ankyrin-B deficiency.
Proceedings of the National Academy of Sciences of the United States of America
Lorenzo DN,Bennett V
Tue Nov 28 00:00:00 EST 2017
Cell-autonomous adiposity through increased cell surface GLUT4 due to ankyrin-B deficiency.
Proceedings of the National Academy of Sciences of the United States of America
Lorenzo DN,Bennett V
Tue Nov 28 00:00:00 EST 2017
BAIAP3, a C2 domain-containing Munc13 protein, controls the fate of dense-core vesicles in neuroendocrine cells.
The Journal of cell biology
Zhang X,Jiang S,Mitok KA,Li L,Attie AD,Martin TFJ
Mon Jul 03 00:00:00 EDT 2017
BAIAP3, a C2 domain-containing Munc13 protein, controls the fate of dense-core vesicles in neuroendocrine cells.
The Journal of cell biology
Zhang X,Jiang S,Mitok KA,Li L,Attie AD,Martin TFJ
Mon Jul 03 00:00:00 EDT 2017
Qi-Zhu-Xie-Zhuo-Fang reduces serum uric acid levels and ameliorates renal fibrosis in hyperuricemic nephropathy rats.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Huijuan W,Xiaoxu C,Rui S,Xinghui L,Beibei T,Jianchun M
Sat Jul 01 00:00:00 EDT 2017
Qi-Zhu-Xie-Zhuo-Fang reduces serum uric acid levels and ameliorates renal fibrosis in hyperuricemic nephropathy rats.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Huijuan W,Xiaoxu C,Rui S,Xinghui L,Beibei T,Jianchun M
Sat Jul 01 00:00:00 EDT 2017
NOR1 Suppresses Cancer Stem-Like Cells Properties of Tumor Cells via the Inhibition of the AKT-GSK-3β-Wnt/β-catenin-ALDH1A1 Signal Circuit.
Journal of cellular physiology
Wang W,Yi M,Chen S,Li J,Zhang H,Xiong W,Li G,Li X,Xiang B
Sun Oct 01 00:00:00 EDT 2017
NOR1 Suppresses Cancer Stem-Like Cells Properties of Tumor Cells via the Inhibition of the AKT-GSK-3β-Wnt/β-catenin-ALDH1A1 Signal Circuit.
Journal of cellular physiology
Wang W,Yi M,Chen S,Li J,Zhang H,Xiong W,Li G,Li X,Xiang B
Sun Oct 01 00:00:00 EDT 2017
HIF-1α coordinates epigenetic activation of SIAH1 in hepatocytes in response to nutritional stress.
Biochimica et biophysica acta. Gene regulatory mechanisms
Fan Z,Li Z,Yang Y,Liu S,Guo J,Xu Y
Sun Oct 01 00:00:00 EDT 2017
HIF-1α coordinates epigenetic activation of SIAH1 in hepatocytes in response to nutritional stress.
Biochimica et biophysica acta. Gene regulatory mechanisms
Fan Z,Li Z,Yang Y,Liu S,Guo J,Xu Y
Sun Oct 01 00:00:00 EDT 2017
Long-Term Dietary Supplementation with Selenium-Enriched Yeast Improves Cognitive Impairment, Reverses Synaptic Deficits, and Mitigates Tau Pathology in a Triple Transgenic Mouse Model of Alzheimer's Disease.
Journal of agricultural and food chemistry
Zhang ZH,Wen L,Wu QY,Chen C,Zheng R,Liu Q,Ni JZ,Song GL
Wed Jun 21 00:00:00 EDT 2017
Long-Term Dietary Supplementation with Selenium-Enriched Yeast Improves Cognitive Impairment, Reverses Synaptic Deficits, and Mitigates Tau Pathology in a Triple Transgenic Mouse Model of Alzheimer's Disease.
Journal of agricultural and food chemistry
Zhang ZH,Wen L,Wu QY,Chen C,Zheng R,Liu Q,Ni JZ,Song GL
Wed Jun 21 00:00:00 EDT 2017
Anthocyanins from Black Chokeberry (Aroniamelanocarpa Elliot) Delayed Aging-Related Degenerative Changes of Brain.
Journal of agricultural and food chemistry
Wei J,Zhang G,Zhang X,Xu D,Gao J,Fan J,Zhou Z
Wed Jul 26 00:00:00 EDT 2017
Anthocyanins from Black Chokeberry (Aroniamelanocarpa Elliot) Delayed Aging-Related Degenerative Changes of Brain.
Journal of agricultural and food chemistry
Wei J,Zhang G,Zhang X,Xu D,Gao J,Fan J,Zhou Z
Wed Jul 26 00:00:00 EDT 2017
Downregulation of NAD-Dependent Deacetylase SIRT2 Protects Mouse Brain Against Ischemic Stroke.
Molecular neurobiology
Xie XQ,Zhang P,Tian B,Chen XQ
Wed Nov 01 00:00:00 EDT 2017
Downregulation of NAD-Dependent Deacetylase SIRT2 Protects Mouse Brain Against Ischemic Stroke.
Molecular neurobiology
Xie XQ,Zhang P,Tian B,Chen XQ
Wed Nov 01 00:00:00 EDT 2017
Satb2 Ablation Impairs Hippocampus-Based Long-Term Spatial Memory and Short-Term Working Memory and Immediate Early Genes (IEGs)-Mediated Hippocampal Synaptic Plasticity.
Molecular neurobiology
Li Y,You QL,Zhang SR,Huang WY,Zou WJ,Jie W,Li SJ,Liu JH,Lv CY,Cong J,Hu YY,Gao TM,Li JM
Tue Apr 18 00:00:00 EDT 2017
Satb2 Ablation Impairs Hippocampus-Based Long-Term Spatial Memory and Short-Term Working Memory and Immediate Early Genes (IEGs)-Mediated Hippocampal Synaptic Plasticity.
Molecular neurobiology
Li Y,You QL,Zhang SR,Huang WY,Zou WJ,Jie W,Li SJ,Liu JH,Lv CY,Cong J,Hu YY,Gao TM,Li JM
Tue Apr 18 00:00:00 EDT 2017
Motile hepatocellular carcinoma cells preferentially secret sugar metabolism regulatory proteins via exosomes.
Proteomics
Zhang J,Lu S,Zhou Y,Meng K,Chen Z,Cui Y,Shi Y,Wang T,He QY
Sat Jul 01 00:00:00 EDT 2017
Motile hepatocellular carcinoma cells preferentially secret sugar metabolism regulatory proteins via exosomes.
Proteomics
Zhang J,Lu S,Zhou Y,Meng K,Chen Z,Cui Y,Shi Y,Wang T,He QY
Sat Jul 01 00:00:00 EDT 2017
MicroRNA let-7g possesses a therapeutic potential for peripheral artery disease.
Journal of cellular and molecular medicine
Hsu PY,Hsi E,Wang TM,Lin RT,Liao YC,Juo SH
Wed Mar 01 00:00:00 EST 2017
MicroRNA let-7g possesses a therapeutic potential for peripheral artery disease.
Journal of cellular and molecular medicine
Hsu PY,Hsi E,Wang TM,Lin RT,Liao YC,Juo SH
Wed Mar 01 00:00:00 EST 2017
Role of RAS/Wnt/β-catenin axis activation in the pathogenesis of podocyte injury and tubulo-interstitial nephropathy.
Chemico-biological interactions
Chen L,Chen DQ,Wang M,Liu D,Chen H,Dou F,Vaziri ND,Zhao YY
Tue Aug 01 00:00:00 EDT 2017
Role of RAS/Wnt/β-catenin axis activation in the pathogenesis of podocyte injury and tubulo-interstitial nephropathy.
Chemico-biological interactions
Chen L,Chen DQ,Wang M,Liu D,Chen H,Dou F,Vaziri ND,Zhao YY
Tue Aug 01 00:00:00 EDT 2017
Overexpression of Rsf-1 correlates with poor survival and promotes invasion in non-small cell lung cancer.
Virchows Archiv : an international journal of pathology
Zhang X,Fu L,Xue D,Zhang X,Hao F,Xie L,He J,Gai J,Liu Y,Xu H,Li Q,Wang E
Mon May 01 00:00:00 EDT 2017
Overexpression of Rsf-1 correlates with poor survival and promotes invasion in non-small cell lung cancer.
Virchows Archiv : an international journal of pathology
Zhang X,Fu L,Xue D,Zhang X,Hao F,Xie L,He J,Gai J,Liu Y,Xu H,Li Q,Wang E
Mon May 01 00:00:00 EDT 2017
miR-34b Modulates Skeletal Muscle Cell Proliferation and Differentiation.
Journal of cellular biochemistry
Tang Z,Qiu H,Luo L,Liu N,Zhong J,Kang K,Gou D
Fri Dec 01 00:00:00 EST 2017
miR-34b Modulates Skeletal Muscle Cell Proliferation and Differentiation.
Journal of cellular biochemistry
Tang Z,Qiu H,Luo L,Liu N,Zhong J,Kang K,Gou D
Fri Dec 01 00:00:00 EST 2017
Histone Methyltransferase SETDB1 Promotes the Progression of Colorectal Cancer by Inhibiting the Expression of TP53.
Journal of Cancer
Chen K,Zhang F,Ding J,Liang Y,Zhan Z,Zhan Y,Chen LH,Ding Y
Thu Mar 17 00:00:00 EDT 2022
Histone Methyltransferase SETDB1 Promotes the Progression of Colorectal Cancer by Inhibiting the Expression of TP53.
Journal of Cancer
Chen K,Zhang F,Ding J,Liang Y,Zhan Z,Zhan Y,Chen LH,Ding Y
Thu Mar 17 00:00:00 EDT 2022
Neuroprotective Effects of Lycium barbarum Polysaccharide on Focal Cerebral Ischemic Injury in Mice.
Neurochemical research
Zhao P,Zhou R,Zhu XY,Liu G,Zhao YP,Ma PS,Wu W,Niu Y,Sun T,Li YX,Yu JQ,Qian ZM
Sun Oct 01 00:00:00 EDT 2017
Neuroprotective Effects of Lycium barbarum Polysaccharide on Focal Cerebral Ischemic Injury in Mice.
Neurochemical research
Zhao P,Zhou R,Zhu XY,Liu G,Zhao YP,Ma PS,Wu W,Niu Y,Sun T,Li YX,Yu JQ,Qian ZM
Sun Oct 01 00:00:00 EDT 2017
Knockdown of homeobox containing 1 increases the radiosensitivity of cervical cancer cells through telomere shortening.
Oncology reports
Zhou S,Xiao Y,Zhuang Y,Liu Y,Zhao H,Yang H,Xie C,Zhou F,Zhou Y
Sat Jul 01 00:00:00 EDT 2017
Knockdown of homeobox containing 1 increases the radiosensitivity of cervical cancer cells through telomere shortening.
Oncology reports
Zhou S,Xiao Y,Zhuang Y,Liu Y,Zhao H,Yang H,Xie C,Zhou F,Zhou Y
Sat Jul 01 00:00:00 EDT 2017
Anthocyanins Delay Ageing-Related Degenerative Changes in the Liver.
Plant foods for human nutrition (Dordrecht, Netherlands)
Wei J,Zhang G,Zhang X,Xu D,Gao J,Fan J
Fri Dec 01 00:00:00 EST 2017
Anthocyanins Delay Ageing-Related Degenerative Changes in the Liver.
Plant foods for human nutrition (Dordrecht, Netherlands)
Wei J,Zhang G,Zhang X,Xu D,Gao J,Fan J
Fri Dec 01 00:00:00 EST 2017
Non-hemolytic enterotoxin of Bacillus cereus induces apoptosis in Vero cells.
Cellular microbiology
Liu X,Ding S,Shi P,Dietrich R,Märtlbauer E,Zhu K
Sat Apr 01 00:00:00 EDT 2017
Non-hemolytic enterotoxin of Bacillus cereus induces apoptosis in Vero cells.
Cellular microbiology
Liu X,Ding S,Shi P,Dietrich R,Märtlbauer E,Zhu K
Sat Apr 01 00:00:00 EDT 2017
Analysis of gene expression and functional characterization of XPR1: a pathogenic gene for primary familial brain calcification.
Cell and tissue research
Yao XP,Zhao M,Wang C,Guo XX,Su HZ,Dong EL,Chen HT,Lai JH,Liu YB,Wang N,Chen WJ
Wed Nov 01 00:00:00 EDT 2017
Analysis of gene expression and functional characterization of XPR1: a pathogenic gene for primary familial brain calcification.
Cell and tissue research
Yao XP,Zhao M,Wang C,Guo XX,Su HZ,Dong EL,Chen HT,Lai JH,Liu YB,Wang N,Chen WJ
Wed Nov 01 00:00:00 EDT 2017
HMGB1 binding heptamer peptide improves survival and ameliorates brain injury in rats after cardiac arrest and cardiopulmonary resuscitation.
Neuroscience
Shi X,Li M,Huang K,Zhou S,Hu Y,Pan S,Gu Y
Sat Sep 30 00:00:00 EDT 2017
HMGB1 binding heptamer peptide improves survival and ameliorates brain injury in rats after cardiac arrest and cardiopulmonary resuscitation.
Neuroscience
Shi X,Li M,Huang K,Zhou S,Hu Y,Pan S,Gu Y
Sat Sep 30 00:00:00 EDT 2017
HMGB1 binding heptamer peptide improves survival and ameliorates brain injury in rats after cardiac arrest and cardiopulmonary resuscitation.
Neuroscience
Shi X,Li M,Huang K,Zhou S,Hu Y,Pan S,Gu Y
Sat Sep 30 00:00:00 EDT 2017
Vav1 expression is increased in esophageal squamous cell carcinoma and indicates poor prognosis.
Biochemical and biophysical research communications
Zhu X,Jin H,Xia Z,Wu X,Yang M,Zhang H,Shang X,Cheng R,Zhan Z,Yu Z
Sat Apr 29 00:00:00 EDT 2017
Vav1 expression is increased in esophageal squamous cell carcinoma and indicates poor prognosis.
Biochemical and biophysical research communications
Zhu X,Jin H,Xia Z,Wu X,Yang M,Zhang H,Shang X,Cheng R,Zhan Z,Yu Z
Sat Apr 29 00:00:00 EDT 2017
Atorvastatin prevents Angiotensin II induced myocardial hypertrophy in vitro via CCAAT/enhancer-binding protein β.
Biochemical and biophysical research communications
Chen Y,Yu S,Zhang N,Li Y,Chen S,Chang Y,Sun G,Sun Y
Sat Apr 29 00:00:00 EDT 2017
Atorvastatin prevents Angiotensin II induced myocardial hypertrophy in vitro via CCAAT/enhancer-binding protein β.
Biochemical and biophysical research communications
Chen Y,Yu S,Zhang N,Li Y,Chen S,Chang Y,Sun G,Sun Y
Sat Apr 29 00:00:00 EDT 2017
Stress stimuli induce cancer-stemness gene expression via Sp1 activation leading to therapeutic resistance in glioblastoma.
Biochemical and biophysical research communications
Chang KY,Huang CT,Hsu TI,Hsu CC,Liu JJ,Chuang CK,Hung JJ,Chang WC,Tsai KK,Chuang JY
Sat Nov 04 00:00:00 EDT 2017
Stress stimuli induce cancer-stemness gene expression via Sp1 activation leading to therapeutic resistance in glioblastoma.
Biochemical and biophysical research communications
Chang KY,Huang CT,Hsu TI,Hsu CC,Liu JJ,Chuang CK,Hung JJ,Chang WC,Tsai KK,Chuang JY
Sat Nov 04 00:00:00 EDT 2017
MiR-466b-1-3p regulates P-glycoprotein expression in rat cerebral microvascular endothelial cells.
Neuroscience letters
Yang X,Ren W,Shao Y,Chen Y
Mon Apr 03 00:00:00 EDT 2017
MiR-466b-1-3p regulates P-glycoprotein expression in rat cerebral microvascular endothelial cells.
Neuroscience letters
Yang X,Ren W,Shao Y,Chen Y
Mon Apr 03 00:00:00 EDT 2017
Hydrogen sulfide generation from l-cysteine in the human glioblastoma-astrocytoma U-87 MG and neuroblastoma SHSY5Y cell lines.
Acta biochimica Polonica
Bronowicka-Adamska P,Bentke A,Wróbel M
Mon Apr 17 00:00:00 EDT 2017
Hydrogen sulfide generation from l-cysteine in the human glioblastoma-astrocytoma U-87 MG and neuroblastoma SHSY5Y cell lines.
Acta biochimica Polonica
Bronowicka-Adamska P,Bentke A,Wróbel M
Mon Apr 17 00:00:00 EDT 2017
JOSD1 Negatively Regulates Type-I Interferon Antiviral Activity by Deubiquitinating and Stabilizing SOCS1.
Viral immunology
Wang X,Zhang L,Zhang Y,Zhao P,Qian L,Yuan Y,Liu J,Cheng Q,Xu W,Zuo Y,Guo T,Yu Z,Zheng H
Thu Jun 01 00:00:00 EDT 2017
JOSD1 Negatively Regulates Type-I Interferon Antiviral Activity by Deubiquitinating and Stabilizing SOCS1.
Viral immunology
Wang X,Zhang L,Zhang Y,Zhao P,Qian L,Yuan Y,Liu J,Cheng Q,Xu W,Zuo Y,Guo T,Yu Z,Zheng H
Thu Jun 01 00:00:00 EDT 2017
Thymoquinone Attenuates Brain Injury via an Anti-oxidative Pathway in a Status Epilepticus Rat Model.
Translational neuroscience
Shao YY,Li B,Huang YM,Luo Q,Xie YM,Chen YH
Sun Jan 01 00:00:00 EST 2017
Thymoquinone Attenuates Brain Injury via an Anti-oxidative Pathway in a Status Epilepticus Rat Model.
Translational neuroscience
Shao YY,Li B,Huang YM,Luo Q,Xie YM,Chen YH
Sun Jan 01 00:00:00 EST 2017
Loss of the novel mitochondrial protein FAM210B promotes metastasis via PDK4-dependent metabolic reprogramming.
Cell death & disease
Sun S,Liu J,Zhao M,Han Y,Chen P,Mo Q,Wang B,Chen G,Fang Y,Tian Y,Zhou J,Ma D,Gao Q,Wu P
Thu Jun 08 00:00:00 EDT 2017
Loss of the novel mitochondrial protein FAM210B promotes metastasis via PDK4-dependent metabolic reprogramming.
Cell death & disease
Sun S,Liu J,Zhao M,Han Y,Chen P,Mo Q,Wang B,Chen G,Fang Y,Tian Y,Zhou J,Ma D,Gao Q,Wu P
Thu Jun 08 00:00:00 EDT 2017
FAM83D, a microtubule-associated protein, promotes tumor growth and progression of human gastric cancer.
Oncotarget
Huang M,Ma X,Shi H,Hu L,Fan Z,Pang L,Zhu F,Yang X,Xu W,Liu B,Zhu Z,Li C
Tue Sep 26 00:00:00 EDT 2017
FAM83D, a microtubule-associated protein, promotes tumor growth and progression of human gastric cancer.
Oncotarget
Huang M,Ma X,Shi H,Hu L,Fan Z,Pang L,Zhu F,Yang X,Xu W,Liu B,Zhu Z,Li C
Tue Sep 26 00:00:00 EDT 2017
A signature motif in LIM proteins mediates binding to checkpoint proteins and increases tumour radiosensitivity.
Nature communications
Xu X,Fan Z,Liang C,Li L,Wang L,Liang Y,Wu J,Chang S,Yan Z,Lv Z,Fu J,Liu Y,Jin S,Wang T,Hong T,Dong Y,Ding L,Cheng L,Liu R,Fu S,Jiao S,Ye Q
Tue Jan 17 00:00:00 EST 2017
A signature motif in LIM proteins mediates binding to checkpoint proteins and increases tumour radiosensitivity.
Nature communications
Xu X,Fan Z,Liang C,Li L,Wang L,Liang Y,Wu J,Chang S,Yan Z,Lv Z,Fu J,Liu Y,Jin S,Wang T,Hong T,Dong Y,Ding L,Cheng L,Liu R,Fu S,Jiao S,Ye Q
Tue Jan 17 00:00:00 EST 2017
Characteristics of Chinese herbal medicine usage in ischemic heart disease patients among type 2 diabetes and their protection against hydrogen peroxide-mediated apoptosis in H9C2 cardiomyoblasts.
Oncotarget
Tsai FJ,Ho TJ,Cheng CF,Shiao YT,Chien WK,Chen JH,Liu X,Tsang H,Lin TH,Liao CC,Huang SM,Li JP,Lin CW,Lin JG,Lan YC,Liu YH,Hung CH,Lin JC,Lin CC,Lai CH,Liang WM,Lin YJ
Tue Feb 28 00:00:00 EST 2017
Functional analysis reveals that RBM10 mutations contribute to lung adenocarcinoma pathogenesis by deregulating splicing.
Scientific reports
Zhao J,Sun Y,Huang Y,Song F,Huang Z,Bao Y,Zuo J,Saffen D,Shao Z,Liu W,Wang Y
Mon Jan 16 00:00:00 EST 2017
Functional analysis reveals that RBM10 mutations contribute to lung adenocarcinoma pathogenesis by deregulating splicing.
Scientific reports
Zhao J,Sun Y,Huang Y,Song F,Huang Z,Bao Y,Zuo J,Saffen D,Shao Z,Liu W,Wang Y
Mon Jan 16 00:00:00 EST 2017
MiR-30c regulates cisplatin-induced apoptosis of renal tubular epithelial cells by targeting Bnip3L and Hspa5.
Cell death & disease
Du B,Dai XM,Li S,Qi GL,Cao GX,Zhong Y,Yin PD,Yang XS
Thu Aug 10 00:00:00 EDT 2017
MiR-30c regulates cisplatin-induced apoptosis of renal tubular epithelial cells by targeting Bnip3L and Hspa5.
Cell death & disease
Du B,Dai XM,Li S,Qi GL,Cao GX,Zhong Y,Yin PD,Yang XS
Thu Aug 10 00:00:00 EDT 2017
Antisense Oligonucleotide-Mediated Removal of the Polyglutamine Repeat in Spinocerebellar Ataxia Type 3 Mice.
Molecular therapy. Nucleic acids
Toonen LJA,Rigo F,van Attikum H,van Roon-Mom WMC
Fri Sep 15 00:00:00 EDT 2017
Long noncoding RNA CRNDE stabilized by hnRNPUL2 accelerates cell proliferation and migration in colorectal carcinoma via activating Ras/MAPK signaling pathways.
Cell death & disease
Jiang H,Wang Y,Ai M,Wang H,Duan Z,Wang H,Zhao L,Yu J,Ding Y,Wang S
Thu Jun 08 00:00:00 EDT 2017
Long noncoding RNA CRNDE stabilized by hnRNPUL2 accelerates cell proliferation and migration in colorectal carcinoma via activating Ras/MAPK signaling pathways.
Cell death & disease
Jiang H,Wang Y,Ai M,Wang H,Duan Z,Wang H,Zhao L,Yu J,Ding Y,Wang S
Thu Jun 08 00:00:00 EDT 2017
PTPIP51 regulates mouse cardiac ischemia/reperfusion through mediating the mitochondria-SR junction.
Scientific reports
Qiao X,Jia S,Ye J,Fang X,Zhang C,Cao Y,Xu C,Zhao L,Zhu Y,Wang L,Zheng M
Mon Mar 27 00:00:00 EDT 2017
PTPIP51 regulates mouse cardiac ischemia/reperfusion through mediating the mitochondria-SR junction.
Scientific reports
Qiao X,Jia S,Ye J,Fang X,Zhang C,Cao Y,Xu C,Zhao L,Zhu Y,Wang L,Zheng M
Mon Mar 27 00:00:00 EDT 2017
Palladin is a novel microtubule-associated protein responsible for spindle orientation.
Scientific reports
Zhang X,Chen X,Liu J,Xu X,Zhang Y,Ruan Z,Xie Y,Huang Q,Yin T,Chen Z,Chen S
Mon Sep 18 00:00:00 EDT 2017
Palladin is a novel microtubule-associated protein responsible for spindle orientation.
Scientific reports
Zhang X,Chen X,Liu J,Xu X,Zhang Y,Ruan Z,Xie Y,Huang Q,Yin T,Chen Z,Chen S
Mon Sep 18 00:00:00 EDT 2017
PKC θ-mediated Ca2+/NF-AT signalling pathway may be involved in T-cell immunosuppression in coal-burning arsenic-poisoned population.
Environmental toxicology and pharmacology
Zeng Q,Luo P,Gu J,Liang B,Liu Q,Zhang A
Sun Oct 01 00:00:00 EDT 2017
BRAFV600E and RET/PTC Promote Proliferation and Migration of Papillary Thyroid Carcinoma Cells In Vitro by Regulating Nuclear Factor-κB.
Medical science monitor : international medical journal of experimental and clinical research
Zhou D,Li Z,Bai X
Wed Nov 08 00:00:00 EST 2017
BRAFV600E and RET/PTC Promote Proliferation and Migration of Papillary Thyroid Carcinoma Cells In Vitro by Regulating Nuclear Factor-κB.
Medical science monitor : international medical journal of experimental and clinical research
Zhou D,Li Z,Bai X
Wed Nov 08 00:00:00 EST 2017
Specificity protein 1-modulated superoxide dismutase 2 enhances temozolomide resistance in glioblastoma, which is independent of O6-methylguanine-DNA methyltransferase.
Redox biology
Chang KY,Hsu TI,Hsu CC,Tsai SY,Liu JJ,Chou SW,Liu MS,Liou JP,Ko CY,Chen KY,Hung JJ,Chang WC,Chuang CK,Kao TJ,Chuang JY
Sun Oct 01 00:00:00 EDT 2017
Specificity protein 1-modulated superoxide dismutase 2 enhances temozolomide resistance in glioblastoma, which is independent of O6-methylguanine-DNA methyltransferase.
Redox biology
Chang KY,Hsu TI,Hsu CC,Tsai SY,Liu JJ,Chou SW,Liu MS,Liou JP,Ko CY,Chen KY,Hung JJ,Chang WC,Chuang CK,Kao TJ,Chuang JY
Sun Oct 01 00:00:00 EDT 2017
Overexpression of WDFY2 inhibits prostate cancer cell growth and migration via inactivation of Akt pathway.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine
Wang J,Chen X,Tong S,Zhou H,Sun J,Gou Y,Wu F,Hu J,Xu J,Ding G
Thu Jun 01 00:00:00 EDT 2017
Overexpression of WDFY2 inhibits prostate cancer cell growth and migration via inactivation of Akt pathway.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine
Wang J,Chen X,Tong S,Zhou H,Sun J,Gou Y,Wu F,Hu J,Xu J,Ding G
Thu Jun 01 00:00:00 EDT 2017
Rabies Virus Infection Induces Microtubule Depolymerization to Facilitate Viral RNA Synthesis by Upregulating HDAC6.
Frontiers in cellular and infection microbiology
Zan J,Liu S,Sun DN,Mo KK,Yan Y,Liu J,Hu BL,Gu JY,Liao M,Zhou JY
Wed Dec 27 00:00:00 EST 2017
Rabies Virus Infection Induces Microtubule Depolymerization to Facilitate Viral RNA Synthesis by Upregulating HDAC6.
Frontiers in cellular and infection microbiology
Zan J,Liu S,Sun DN,Mo KK,Yan Y,Liu J,Hu BL,Gu JY,Liao M,Zhou JY
Wed Dec 27 00:00:00 EST 2017
Rabies Virus Infection Induces Microtubule Depolymerization to Facilitate Viral RNA Synthesis by Upregulating HDAC6.
Frontiers in cellular and infection microbiology
Zan J,Liu S,Sun DN,Mo KK,Yan Y,Liu J,Hu BL,Gu JY,Liao M,Zhou JY
Wed Dec 27 00:00:00 EST 2017
Rabies Virus Infection Induces Microtubule Depolymerization to Facilitate Viral RNA Synthesis by Upregulating HDAC6.
Frontiers in cellular and infection microbiology
Zan J,Liu S,Sun DN,Mo KK,Yan Y,Liu J,Hu BL,Gu JY,Liao M,Zhou JY
Wed Dec 27 00:00:00 EST 2017
HRD1 prevents apoptosis in renal tubular epithelial cells by mediating eIF2α ubiquitylation and degradation.
Cell death & disease
Huang Y,Sun Y,Cao Y,Sun H,Li M,You H,Su D,Li Y,Liang X
Mon Dec 11 00:00:00 EST 2017
HRD1 prevents apoptosis in renal tubular epithelial cells by mediating eIF2α ubiquitylation and degradation.
Cell death & disease
Huang Y,Sun Y,Cao Y,Sun H,Li M,You H,Su D,Li Y,Liang X
Mon Dec 11 00:00:00 EST 2017
2-Acetylamino-3-[4-(2-acetylamino-2-carboxyethylsulfanylcarbonylamino) phenyl carbamoylsulfanyl] propionic acid, a glutathione reductase inhibitor, induces G2/M cell cycle arrest through generation of thiol oxidative stress in human esophageal cancer cells.
Oncotarget
Li X,Jiang Z,Feng J,Zhang X,Wu J,Chen W
Fri Sep 22 00:00:00 EDT 2017
2-Acetylamino-3-[4-(2-acetylamino-2-carboxyethylsulfanylcarbonylamino) phenyl carbamoylsulfanyl] propionic acid, a glutathione reductase inhibitor, induces G2/M cell cycle arrest through generation of thiol oxidative stress in human esophageal cancer cells.
Oncotarget
Li X,Jiang Z,Feng J,Zhang X,Wu J,Chen W
Fri Sep 22 00:00:00 EDT 2017
2-Acetylamino-3-[4-(2-acetylamino-2-carboxyethylsulfanylcarbonylamino) phenyl carbamoylsulfanyl] propionic acid, a glutathione reductase inhibitor, induces G2/M cell cycle arrest through generation of thiol oxidative stress in human esophageal cancer cells.
Oncotarget
Li X,Jiang Z,Feng J,Zhang X,Wu J,Chen W
Fri Sep 22 00:00:00 EDT 2017
2-Acetylamino-3-[4-(2-acetylamino-2-carboxyethylsulfanylcarbonylamino) phenyl carbamoylsulfanyl] propionic acid, a glutathione reductase inhibitor, induces G2/M cell cycle arrest through generation of thiol oxidative stress in human esophageal cancer cells.
Oncotarget
Li X,Jiang Z,Feng J,Zhang X,Wu J,Chen W
Fri Sep 22 00:00:00 EDT 2017
Adipose-derived mesenchymal stem cells promote osteosarcoma proliferation and metastasis by activating the STAT3 pathway.
Oncotarget
Wang Y,Chu Y,Yue B,Ma X,Zhang G,Xiang H,Liu Y,Wang T,Wu X,Chen B
Tue Apr 04 00:00:00 EDT 2017
Adipose-derived mesenchymal stem cells promote osteosarcoma proliferation and metastasis by activating the STAT3 pathway.
Oncotarget
Wang Y,Chu Y,Yue B,Ma X,Zhang G,Xiang H,Liu Y,Wang T,Wu X,Chen B
Tue Apr 04 00:00:00 EDT 2017
Inhibition of IL-8-mediated endothelial adhesion, VSMCs proliferation and migration by siRNA-TMEM98 suggests TMEM98's emerging role in atherosclerosis.
Oncotarget
Lv G,Zhu H,Li C,Wang J,Zhao D,Li S,Ma L,Sun G,Li F,Zhao Y,Gao Y
Fri Oct 20 00:00:00 EDT 2017
Inhibition of IL-8-mediated endothelial adhesion, VSMCs proliferation and migration by siRNA-TMEM98 suggests TMEM98's emerging role in atherosclerosis.
Oncotarget
Lv G,Zhu H,Li C,Wang J,Zhao D,Li S,Ma L,Sun G,Li F,Zhao Y,Gao Y
Fri Oct 20 00:00:00 EDT 2017
A critical role of Hrd1 in the regulation of optineurin degradation and aggresome formation.
Human molecular genetics
Mao J,Xia Q,Liu C,Ying Z,Wang H,Wang G
Mon May 15 00:00:00 EDT 2017
MiR-130a regulates neurite outgrowth and dendritic spine density by targeting MeCP2.
Protein & cell
Zhang Y,Chen M,Qiu Z,Hu K,McGee W,Chen X,Liu J,Zhu L,Wu JY
Fri Jul 01 00:00:00 EDT 2016
MiR-130a regulates neurite outgrowth and dendritic spine density by targeting MeCP2.
Protein & cell
Zhang Y,Chen M,Qiu Z,Hu K,McGee W,Chen X,Liu J,Zhu L,Wu JY
Fri Jul 01 00:00:00 EDT 2016
The Ubiquitin-like Protein FAT10 Stabilizes eEF1A1 Expression to Promote Tumor Proliferation in a Complex Manner.
Cancer research
Liu X,Chen L,Ge J,Yan C,Huang Z,Hu J,Wen C,Li M,Huang D,Qiu Y,Hao H,Yuan R,Lei J,Yu X,Shao J
Mon Aug 15 00:00:00 EDT 2016
The Ubiquitin-like Protein FAT10 Stabilizes eEF1A1 Expression to Promote Tumor Proliferation in a Complex Manner.
Cancer research
Liu X,Chen L,Ge J,Yan C,Huang Z,Hu J,Wen C,Li M,Huang D,Qiu Y,Hao H,Yuan R,Lei J,Yu X,Shao J
Mon Aug 15 00:00:00 EDT 2016
miR-1301 promotes prostate cancer proliferation through directly targeting PPP2R2C.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Bi D,Ning H,Liu S,Que X,Ding K
Fri Jul 01 00:00:00 EDT 2016
miR-1301 promotes prostate cancer proliferation through directly targeting PPP2R2C.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Bi D,Ning H,Liu S,Que X,Ding K
Fri Jul 01 00:00:00 EDT 2016
Anxa5 mediates the in vitro malignant behaviours of murine hepatocarcinoma Hca-F cells with high lymph node metastasis potential preferentially via ERK2/p-ERK2/c-Jun/p-c-Jun(Ser73) and E-cadherin.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Sun X,Wei B,Liu S,Guo C,Wu N,Liu Q,Sun MZ
Thu Dec 01 00:00:00 EST 2016
Anxa5 mediates the in vitro malignant behaviours of murine hepatocarcinoma Hca-F cells with high lymph node metastasis potential preferentially via ERK2/p-ERK2/c-Jun/p-c-Jun(Ser73) and E-cadherin.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Sun X,Wei B,Liu S,Guo C,Wu N,Liu Q,Sun MZ
Thu Dec 01 00:00:00 EST 2016
Achaete-Scute Homolog 1 Expression Controls Cellular Differentiation of Neuroblastoma.
Frontiers in molecular neuroscience
Kasim M,Heß V,Scholz H,Persson PB,Fähling M
Thu Oct 01 00:00:00 EDT 2020
Achaete-Scute Homolog 1 Expression Controls Cellular Differentiation of Neuroblastoma.
Frontiers in molecular neuroscience
Kasim M,Heß V,Scholz H,Persson PB,Fähling M
Thu Oct 01 00:00:00 EDT 2020
CFIm25 regulates glutaminase alternative terminal exon definition to modulate miR-23 function.
RNA (New York, N.Y.)
Masamha CP,Xia Z,Peart N,Collum S,Li W,Wagner EJ,Shyu AB
Wed Jun 01 00:00:00 EDT 2016
Renalase attenuates hypertension, renal injury and cardiac remodelling in rats with subtotal nephrectomy.
Journal of cellular and molecular medicine
Yin J,Lu Z,Wang F,Jiang Z,Lu L,Miao N,Wang N
Wed Jun 01 00:00:00 EDT 2016
Renalase attenuates hypertension, renal injury and cardiac remodelling in rats with subtotal nephrectomy.
Journal of cellular and molecular medicine
Yin J,Lu Z,Wang F,Jiang Z,Lu L,Miao N,Wang N
Wed Jun 01 00:00:00 EDT 2016
Go is required for the release of IL-8 and TNF-α, but not degranulation in human mast cells.
European journal of pharmacology
Yu Y,Huang Z,Mao Z,Zhang Y,Jin M,Chen W,Zhang W,Yu B,Zhang W,Alaster Lau HY
Sun Jun 05 00:00:00 EDT 2016
Go is required for the release of IL-8 and TNF-α, but not degranulation in human mast cells.
European journal of pharmacology
Yu Y,Huang Z,Mao Z,Zhang Y,Jin M,Chen W,Zhang W,Yu B,Zhang W,Alaster Lau HY
Sun Jun 05 00:00:00 EDT 2016
Protective Effects of Thymoquinone Against Convulsant Activity Induced by Lithium-Pilocarpine in a model of Status Epilepticus.
Neurochemical research
Shao Y,Feng Y,Xie Y,Luo Q,Chen L,Li B,Chen Y
Thu Dec 01 00:00:00 EST 2016
Protective Effects of Thymoquinone Against Convulsant Activity Induced by Lithium-Pilocarpine in a model of Status Epilepticus.
Neurochemical research
Shao Y,Feng Y,Xie Y,Luo Q,Chen L,Li B,Chen Y
Thu Dec 01 00:00:00 EST 2016
Characterization of Brain-Penetrant Pyrimidine-Containing Molecules with Differential Microtubule-Stabilizing Activities Developed as Potential Therapeutic Agents for Alzheimer's Disease and Related Tauopathies.
The Journal of pharmacology and experimental therapeutics
Kovalevich J,Cornec AS,Yao Y,James M,Crowe A,Lee VM,Trojanowski JQ,Smith AB,Ballatore C,Brunden KR
Sun May 01 00:00:00 EDT 2016
Characterization of Brain-Penetrant Pyrimidine-Containing Molecules with Differential Microtubule-Stabilizing Activities Developed as Potential Therapeutic Agents for Alzheimer's Disease and Related Tauopathies.
The Journal of pharmacology and experimental therapeutics
Kovalevich J,Cornec AS,Yao Y,James M,Crowe A,Lee VM,Trojanowski JQ,Smith AB,Ballatore C,Brunden KR
Sun May 01 00:00:00 EDT 2016
High-dose irradiation in combination with toll-like receptor 9 agonist CpG oligodeoxynucleotide 7909 downregulates PD-L1 expression via the NF-κB signaling pathway in non-small cell lung cancer cells.
OncoTargets and therapy
Chen X,Zhang Q,Luo Y,Gao C,Zhuang X,Xu G,Qiao T
Thu Oct 01 00:00:00 EDT 2020
High-dose irradiation in combination with toll-like receptor 9 agonist CpG oligodeoxynucleotide 7909 downregulates PD-L1 expression via the NF-κB signaling pathway in non-small cell lung cancer cells.
OncoTargets and therapy
Chen X,Zhang Q,Luo Y,Gao C,Zhuang X,Xu G,Qiao T
Thu Oct 01 00:00:00 EDT 2020
Tetraiodothyroacetic acid and transthyretin silencing inhibit pro-metastatic effect of L-thyroxin in anoikis-resistant prostate cancer cells through regulation of MAPK/ERK pathway.
Experimental cell research
Zhang P,Chen L,Song Y,Li X,Sun Y,Xiao Y,Xing Y
Sat Oct 01 00:00:00 EDT 2016
Tetraiodothyroacetic acid and transthyretin silencing inhibit pro-metastatic effect of L-thyroxin in anoikis-resistant prostate cancer cells through regulation of MAPK/ERK pathway.
Experimental cell research
Zhang P,Chen L,Song Y,Li X,Sun Y,Xiao Y,Xing Y
Sat Oct 01 00:00:00 EDT 2016
MicroRNA-106a regulates phosphatase and tensin homologue expression and promotes the proliferation and invasion of ovarian cancer cells.
Oncology reports
Chen L,Zhang F,Sheng XG,Zhang SQ,Chen YT,Liu BW
Sat Oct 01 00:00:00 EDT 2016
MicroRNA-106a regulates phosphatase and tensin homologue expression and promotes the proliferation and invasion of ovarian cancer cells.
Oncology reports
Chen L,Zhang F,Sheng XG,Zhang SQ,Chen YT,Liu BW
Sat Oct 01 00:00:00 EDT 2016
Etomidate inhibits nuclear factor-κB through decreased expression of glucocorticoid receptor in septic rats.
Molecular medicine reports
Zhang Y,Li RM,Wang C,Liu N,Lv S,Xiong JY
Thu Dec 01 00:00:00 EST 2016
Etomidate inhibits nuclear factor-κB through decreased expression of glucocorticoid receptor in septic rats.
Molecular medicine reports
Zhang Y,Li RM,Wang C,Liu N,Lv S,Xiong JY
Thu Dec 01 00:00:00 EST 2016
Rabies virus inactivates cofilin to facilitate viral budding and release.
Biochemical and biophysical research communications
Zan J,An ST,Mo KK,Zhou JW,Liu J,Wang HL,Yan Y,Liao M,Zhou JY
Fri Sep 02 00:00:00 EDT 2016
Rabies virus inactivates cofilin to facilitate viral budding and release.
Biochemical and biophysical research communications
Zan J,An ST,Mo KK,Zhou JW,Liu J,Wang HL,Yan Y,Liao M,Zhou JY
Fri Sep 02 00:00:00 EDT 2016
Niacin receptor GPR109A inhibits insulin secretion and is down-regulated in type 2 diabetic islet beta-cells.
General and comparative endocrinology
Wang N,Guo DY,Tian X,Lin HP,Li YP,Chen SJ,Fu YC,Xu WC,Wei CJ
Sat Oct 01 00:00:00 EDT 2016
Niacin receptor GPR109A inhibits insulin secretion and is down-regulated in type 2 diabetic islet beta-cells.
General and comparative endocrinology
Wang N,Guo DY,Tian X,Lin HP,Li YP,Chen SJ,Fu YC,Xu WC,Wei CJ
Sat Oct 01 00:00:00 EDT 2016
Toll-like receptor 2-mediated MAPKs and NF-κB activation requires the GNAO1-dependent pathway in human mast cells.
Integrative biology : quantitative biosciences from nano to macro
Jin M,Yu B,Zhang W,Zhang W,Xiao Z,Mao Z,Lai Y,Lin D,Ma Q,Pan E,Zhang Y,Yu Y
Mon Sep 12 00:00:00 EDT 2016
Toll-like receptor 2-mediated MAPKs and NF-κB activation requires the GNAO1-dependent pathway in human mast cells.
Integrative biology : quantitative biosciences from nano to macro
Jin M,Yu B,Zhang W,Zhang W,Xiao Z,Mao Z,Lai Y,Lin D,Ma Q,Pan E,Zhang Y,Yu Y
Mon Sep 12 00:00:00 EDT 2016
Spatially coordinated dynamic gene transcription in living pituitary tissue.
eLife
Featherstone K,Hey K,Momiji H,McNamara AV,Patist AL,Woodburn J,Spiller DG,Christian HC,McNeilly AS,Mullins JJ,Finkenstädt BF,Rand DA,White MR,Davis JR
Mon Feb 01 00:00:00 EST 2016
Spatially coordinated dynamic gene transcription in living pituitary tissue.
eLife
Featherstone K,Hey K,Momiji H,McNamara AV,Patist AL,Woodburn J,Spiller DG,Christian HC,McNeilly AS,Mullins JJ,Finkenstädt BF,Rand DA,White MR,Davis JR
Mon Feb 01 00:00:00 EST 2016
Cell Autonomous and Nonautonomous Function of CUL4B in Mouse Spermatogenesis.
The Journal of biological chemistry
Yin Y,Liu L,Yang C,Lin C,Veith GM,Wang C,Sutovsky P,Zhou P,Ma L
Fri Mar 25 00:00:00 EDT 2016
Cell Autonomous and Nonautonomous Function of CUL4B in Mouse Spermatogenesis.
The Journal of biological chemistry
Yin Y,Liu L,Yang C,Lin C,Veith GM,Wang C,Sutovsky P,Zhou P,Ma L
Fri Mar 25 00:00:00 EDT 2016
CGP57380 enhances efficacy of RAD001 in non-small cell lung cancer through abrogating mTOR inhibition-induced phosphorylation of eIF4E and activating mitochondrial apoptotic pathway.
Oncotarget
Wen Q,Wang W,Luo J,Chu S,Chen L,Xu L,Zang H,Alnemah MM,Ma J,Fan S
Tue May 10 00:00:00 EDT 2016
CGP57380 enhances efficacy of RAD001 in non-small cell lung cancer through abrogating mTOR inhibition-induced phosphorylation of eIF4E and activating mitochondrial apoptotic pathway.
Oncotarget
Wen Q,Wang W,Luo J,Chu S,Chen L,Xu L,Zang H,Alnemah MM,Ma J,Fan S
Tue May 10 00:00:00 EDT 2016
ZBTB20 is required for anterior pituitary development and lactotrope specification.
Nature communications
Cao D,Ma X,Cai J,Luan J,Liu AJ,Yang R,Cao Y,Zhu X,Zhang H,Chen YX,Shi Y,Shi GX,Zou D,Cao X,Grusby MJ,Xie Z,Zhang WJ
Fri Apr 15 00:00:00 EDT 2016
ZBTB20 is required for anterior pituitary development and lactotrope specification.
Nature communications
Cao D,Ma X,Cai J,Luan J,Liu AJ,Yang R,Cao Y,Zhu X,Zhang H,Chen YX,Shi Y,Shi GX,Zou D,Cao X,Grusby MJ,Xie Z,Zhang WJ
Fri Apr 15 00:00:00 EDT 2016
Inhibition of histone deacetylases induces formation of multipolar spindles and subsequent p53-dependent apoptosis in nasopharyngeal carcinoma cells.
Oncotarget
Yan M,Qian YM,Yue CF,Wang ZF,Wang BC,Zhang W,Zheng FM,Liu Q
Tue Jul 12 00:00:00 EDT 2016
Inhibition of histone deacetylases induces formation of multipolar spindles and subsequent p53-dependent apoptosis in nasopharyngeal carcinoma cells.
Oncotarget
Yan M,Qian YM,Yue CF,Wang ZF,Wang BC,Zhang W,Zheng FM,Liu Q
Tue Jul 12 00:00:00 EDT 2016
Premutation in the Fragile X Mental Retardation 1 (FMR1) Gene Affects Maternal Zn-milk and Perinatal Brain Bioenergetics and Scaffolding.
Frontiers in neuroscience
Napoli E,Ross-Inta C,Song G,Wong S,Hagerman R,Gane LW,Smilowitz JT,Tassone F,Giulivi C
Thu May 05 00:00:00 EDT 2016
Premutation in the Fragile X Mental Retardation 1 (FMR1) Gene Affects Maternal Zn-milk and Perinatal Brain Bioenergetics and Scaffolding.
Frontiers in neuroscience
Napoli E,Ross-Inta C,Song G,Wong S,Hagerman R,Gane LW,Smilowitz JT,Tassone F,Giulivi C
Thu May 05 00:00:00 EDT 2016
Gp130-mediated STAT3 activation by S-propargyl-cysteine, an endogenous hydrogen sulfide initiator, prevents doxorubicin-induced cardiotoxicity.
Cell death & disease
Wu J,Guo W,Lin SZ,Wang ZJ,Kan JT,Chen SY,Zhu YZ
Thu Aug 18 00:00:00 EDT 2016
Gp130-mediated STAT3 activation by S-propargyl-cysteine, an endogenous hydrogen sulfide initiator, prevents doxorubicin-induced cardiotoxicity.
Cell death & disease
Wu J,Guo W,Lin SZ,Wang ZJ,Kan JT,Chen SY,Zhu YZ
Thu Aug 18 00:00:00 EDT 2016
Sulforaphane induces apoptosis and inhibits invasion in U251MG glioblastoma cells.
SpringerPlus
Zhang Z,Li C,Shang L,Zhang Y,Zou R,Zhan Y,Bi B
Wed Mar 30 00:00:00 EDT 2016
PfIRR Interacts with HrIGF-I and Activates the MAP-kinase and PI3-kinase Signaling Pathways to Regulate Glycogen Metabolism in Pinctada fucata.
Scientific reports
Shi Y,He MX
Thu Feb 25 00:00:00 EST 2016
PfIRR Interacts with HrIGF-I and Activates the MAP-kinase and PI3-kinase Signaling Pathways to Regulate Glycogen Metabolism in Pinctada fucata.
Scientific reports
Shi Y,He MX
Thu Feb 25 00:00:00 EST 2016
Loss-of-function Mutation in PMVK Causes Autosomal Dominant Disseminated Superficial Porokeratosis.
Scientific reports
Wang J,Liu Y,Liu F,Huang C,Han S,Lv Y,Liu CJ,Zhang S,Qin Y,Ling L,Gao M,Yu S,Li C,Huang M,Liao S,Hu X,Lu Z,Liu X,Jiang T,Tang Z,Zhang H,Guo AY,Liu M
Thu Apr 07 00:00:00 EDT 2016
Loss-of-function Mutation in PMVK Causes Autosomal Dominant Disseminated Superficial Porokeratosis.
Scientific reports
Wang J,Liu Y,Liu F,Huang C,Han S,Lv Y,Liu CJ,Zhang S,Qin Y,Ling L,Gao M,Yu S,Li C,Huang M,Liao S,Hu X,Lu Z,Liu X,Jiang T,Tang Z,Zhang H,Guo AY,Liu M
Thu Apr 07 00:00:00 EDT 2016
Deletion of Brg1 causes abnormal hair cell planer polarity, hair cell anchorage, and scar formation in mouse cochlea.
Scientific reports
Jin Y,Ren N,Li S,Fu X,Sun X,Men Y,Xu Z,Zhang J,Xie Y,Xia M,Gao J
Fri Jun 03 00:00:00 EDT 2016
Deletion of Brg1 causes abnormal hair cell planer polarity, hair cell anchorage, and scar formation in mouse cochlea.
Scientific reports
Jin Y,Ren N,Li S,Fu X,Sun X,Men Y,Xu Z,Zhang J,Xie Y,Xia M,Gao J
Fri Jun 03 00:00:00 EDT 2016
Polypeptide N-Acetylgalactosaminyltransferase 13 Contributes to Neurogenesis via Stabilizing the Mucin-type O-Glycoprotein Podoplanin.
The Journal of biological chemistry
Xu Y,Pang W,Lu J,Shan A,Zhang Y
Fri Nov 04 00:00:00 EDT 2016
Polypeptide N-Acetylgalactosaminyltransferase 13 Contributes to Neurogenesis via Stabilizing the Mucin-type O-Glycoprotein Podoplanin.
The Journal of biological chemistry
Xu Y,Pang W,Lu J,Shan A,Zhang Y
Fri Nov 04 00:00:00 EDT 2016
Pdcd4 restrains the self-renewal and white-to-beige transdifferentiation of adipose-derived stem cells.
Cell death & disease
Bai Y,Shang Q,Zhao H,Pan Z,Guo C,Zhang L,Wang Q
Thu Mar 31 00:00:00 EDT 2016
Pdcd4 restrains the self-renewal and white-to-beige transdifferentiation of adipose-derived stem cells.
Cell death & disease
Bai Y,Shang Q,Zhao H,Pan Z,Guo C,Zhang L,Wang Q
Thu Mar 31 00:00:00 EDT 2016
Nogo-B receptor promotes the chemoresistance of human hepatocellular carcinoma via the ubiquitination of p53 protein.
Oncotarget
Dong C,Zhao B,Long F,Liu Y,Liu Z,Li S,Yang X,Sun D,Wang H,Liu Q,Liang R,Li Y,Gao Z,Shao S,Miao QR,Wang L
Tue Feb 23 00:00:00 EST 2016
NLRP3 inflammasome mediates contrast media-induced acute kidney injury by regulating cell apoptosis.
Scientific reports
Shen J,Wang L,Jiang N,Mou S,Zhang M,Gu L,Shao X,Wang Q,Qi C,Li S,Wang W,Che X,Ni Z
Mon Oct 10 00:00:00 EDT 2016
NLRP3 inflammasome mediates contrast media-induced acute kidney injury by regulating cell apoptosis.
Scientific reports
Shen J,Wang L,Jiang N,Mou S,Zhang M,Gu L,Shao X,Wang Q,Qi C,Li S,Wang W,Che X,Ni Z
Mon Oct 10 00:00:00 EDT 2016
miR-935 promotes gastric cancer cell proliferation by targeting SOX7.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Yang M,Cui G,Ding M,Yang W,Liu Y,Dai D,Chen L
Fri Apr 01 00:00:00 EDT 2016
SUMOylated MAFB promotes colorectal cancer tumorigenesis.
Oncotarget
Yang LS,Zhang XJ,Xie YY,Sun XJ,Zhao R,Huang QH
Tue Dec 13 00:00:00 EST 2016
Elaiophylin, a novel autophagy inhibitor, exerts antitumor activity as a single agent in ovarian cancer cells.
Autophagy
Zhao X,Fang Y,Yang Y,Qin Y,Wu P,Wang T,Lai H,Meng L,Wang D,Zheng Z,Lu X,Zhang H,Gao Q,Zhou J,Ma D
Tue Sep 13 00:00:00 EDT 2016
Elaiophylin, a novel autophagy inhibitor, exerts antitumor activity as a single agent in ovarian cancer cells.
Autophagy
Zhao X,Fang Y,Yang Y,Qin Y,Wu P,Wang T,Lai H,Meng L,Wang D,Zheng Z,Lu X,Zhang H,Gao Q,Zhou J,Ma D
Tue Sep 13 00:00:00 EDT 2016
IFN-γ-mediated IRF1/miR-29b feedback loop suppresses colorectal cancer cell growth and metastasis by repressing IGF1.
Cancer letters
Yuan L,Zhou C,Lu Y,Hong M,Zhang Z,Zhang Z,Chang Y,Zhang C,Li X
Wed Apr 01 00:00:00 EDT 2015
IFN-γ-mediated IRF1/miR-29b feedback loop suppresses colorectal cancer cell growth and metastasis by repressing IGF1.
Cancer letters
Yuan L,Zhou C,Lu Y,Hong M,Zhang Z,Zhang Z,Chang Y,Zhang C,Li X
Wed Apr 01 00:00:00 EDT 2015
Proteomic characterization of the nucleolar linker histone H1 interaction network.
Journal of molecular biology
Szerlong HJ,Herman JA,Krause CM,DeLuca JG,Skoultchi A,Winger QA,Prenni JE,Hansen JC
Fri Jun 05 00:00:00 EDT 2015
Proteomic characterization of the nucleolar linker histone H1 interaction network.
Journal of molecular biology
Szerlong HJ,Herman JA,Krause CM,DeLuca JG,Skoultchi A,Winger QA,Prenni JE,Hansen JC
Fri Jun 05 00:00:00 EDT 2015
The role of autophagy in the cytotoxicity induced by recombinant human arginase in laryngeal squamous cell carcinoma.
Applied microbiology and biotechnology
Lin C,Wang Z,Li L,He Y,Fan J,Liu Z,Zhao S,Ju D
Thu Oct 01 00:00:00 EDT 2015
The role of autophagy in the cytotoxicity induced by recombinant human arginase in laryngeal squamous cell carcinoma.
Applied microbiology and biotechnology
Lin C,Wang Z,Li L,He Y,Fan J,Liu Z,Zhao S,Ju D
Thu Oct 01 00:00:00 EDT 2015
Bipartite Nuclear Localization Signal Controls Nuclear Import and DNA-Binding Activity of IFN Regulatory Factor 3.
Journal of immunology (Baltimore, Md. : 1950)
Zhu M,Fang T,Li S,Meng K,Guo D
Wed Jul 01 00:00:00 EDT 2015
Bipartite Nuclear Localization Signal Controls Nuclear Import and DNA-Binding Activity of IFN Regulatory Factor 3.
Journal of immunology (Baltimore, Md. : 1950)
Zhu M,Fang T,Li S,Meng K,Guo D
Wed Jul 01 00:00:00 EDT 2015
Shear-sensitive microRNA-34a modulates flow-dependent regulation of endothelial inflammation.
Journal of cell science
Fan W,Fang R,Wu X,Liu J,Feng M,Dai G,Chen G,Wu G
Thu Jan 01 00:00:00 EST 2015
Shear-sensitive microRNA-34a modulates flow-dependent regulation of endothelial inflammation.
Journal of cell science
Fan W,Fang R,Wu X,Liu J,Feng M,Dai G,Chen G,Wu G
Thu Jan 01 00:00:00 EST 2015
MNRR1 (formerly CHCHD2) is a bi-organellar regulator of mitochondrial metabolism.
Mitochondrion
Aras S,Bai M,Lee I,Springett R,Hüttemann M,Grossman LI
Thu Jan 01 00:00:00 EST 2015
MNRR1 (formerly CHCHD2) is a bi-organellar regulator of mitochondrial metabolism.
Mitochondrion
Aras S,Bai M,Lee I,Springett R,Hüttemann M,Grossman LI
Thu Jan 01 00:00:00 EST 2015
Adipose-derived mesenchymal stem cells promote cell proliferation and invasion of epithelial ovarian cancer.
Experimental cell research
Chu Y,Tang H,Guo Y,Guo J,Huang B,Fang F,Cai J,Wang Z
Thu Sep 10 00:00:00 EDT 2015
Adipose-derived mesenchymal stem cells promote cell proliferation and invasion of epithelial ovarian cancer.
Experimental cell research
Chu Y,Tang H,Guo Y,Guo J,Huang B,Fang F,Cai J,Wang Z
Thu Sep 10 00:00:00 EDT 2015
High expression of transforming acidic coiled coil-containing protein 3 strongly correlates with aggressive characteristics and poor prognosis of gastric cancer.
Oncology reports
Yun M,Rong J,Lin ZR,He YL,Zhang JX,Peng ZW,Tang LQ,Zeng MS,Zhong Q,Ye S
Tue Sep 01 00:00:00 EDT 2015
Pigment Epithelium-Derived Factor Induces Endothelial Barrier Dysfunction via p38/MAPK Phosphorylation.
BioMed research international
He T,Zhao L,Zhang D,Zhang Q,Jia J,Hu J,Huang Y
Wed Aug 17 00:00:00 EDT 2016
Pigment Epithelium-Derived Factor Induces Endothelial Barrier Dysfunction via p38/MAPK Phosphorylation.
BioMed research international
He T,Zhao L,Zhang D,Zhang Q,Jia J,Hu J,Huang Y
Wed Aug 17 00:00:00 EDT 2016
Hypoxia inducible factor-1 mediates expression of miR-322: potential role in proliferation and migration of pulmonary arterial smooth muscle cells.
Scientific reports
Zeng Y,Liu H,Kang K,Wang Z,Hui G,Zhang X,Zhong J,Peng W,Ramchandran R,Raj JU,Gou D
Mon Jul 13 00:00:00 EDT 2015
Hypoxia inducible factor-1 mediates expression of miR-322: potential role in proliferation and migration of pulmonary arterial smooth muscle cells.
Scientific reports
Zeng Y,Liu H,Kang K,Wang Z,Hui G,Zhang X,Zhong J,Peng W,Ramchandran R,Raj JU,Gou D
Mon Jul 13 00:00:00 EDT 2015
P38/MAPK contributes to endothelial barrier dysfunction via MAP4 phosphorylation-dependent microtubule disassembly in inflammation-induced acute lung injury.
Scientific reports
Li L,Hu J,He T,Zhang Q,Yang X,Lan X,Zhang D,Mei H,Chen B,Huang Y
Mon Mar 09 00:00:00 EDT 2015
P38/MAPK contributes to endothelial barrier dysfunction via MAP4 phosphorylation-dependent microtubule disassembly in inflammation-induced acute lung injury.
Scientific reports
Li L,Hu J,He T,Zhang Q,Yang X,Lan X,Zhang D,Mei H,Chen B,Huang Y
Mon Mar 09 00:00:00 EDT 2015
Cytoplasmic hnRNPK interacts with GSK3β and is essential for the osteoclast differentiation.
Scientific reports
Fan X,Xiong H,Wei J,Gao X,Feng Y,Liu X,Zhang G,He QY,Xu J,Liu L
Mon Dec 07 00:00:00 EST 2015
Cytoplasmic hnRNPK interacts with GSK3β and is essential for the osteoclast differentiation.
Scientific reports
Fan X,Xiong H,Wei J,Gao X,Feng Y,Liu X,Zhang G,He QY,Xu J,Liu L
Mon Dec 07 00:00:00 EST 2015
Real Time Monitoring of Inhibition of Adipogenesis and Angiogenesis by (-)-Epigallocatechin-3-Gallate in 3T3-L1 Adipocytes and Human Umbilical Vein Endothelial Cells.
Nutrients
Tang W,Song H,Cai W,Shen X
Tue Oct 27 00:00:00 EDT 2015
Real Time Monitoring of Inhibition of Adipogenesis and Angiogenesis by (-)-Epigallocatechin-3-Gallate in 3T3-L1 Adipocytes and Human Umbilical Vein Endothelial Cells.
Nutrients
Tang W,Song H,Cai W,Shen X
Tue Oct 27 00:00:00 EDT 2015
The role of autophagy in the neurotoxicity of cationic PAMAM dendrimers.
Biomaterials
Wang S,Li Y,Fan J,Wang Z,Zeng X,Sun Y,Song P,Ju D
Fri Aug 01 00:00:00 EDT 2014
The role of autophagy in the neurotoxicity of cationic PAMAM dendrimers.
Biomaterials
Wang S,Li Y,Fan J,Wang Z,Zeng X,Sun Y,Song P,Ju D
Fri Aug 01 00:00:00 EDT 2014
Insulin-like growth factor binding protein 4 enhances cardiomyocytes induction in murine-induced pluripotent stem cells.
Journal of cellular biochemistry
Xue Y,Yan Y,Gong H,Fang B,Zhou Y,Ding Z,Yin P,Zhang G,Ye Y,Yang C,Ge J,Zou Y
Mon Sep 01 00:00:00 EDT 2014
Insulin-like growth factor binding protein 4 enhances cardiomyocytes induction in murine-induced pluripotent stem cells.
Journal of cellular biochemistry
Xue Y,Yan Y,Gong H,Fang B,Zhou Y,Ding Z,Yin P,Zhang G,Ye Y,Yang C,Ge J,Zou Y
Mon Sep 01 00:00:00 EDT 2014
Probing the cell death signaling pathway of HepG2 cell line induced by copper-1,10-phenanthroline complex.
Biometals : an international journal on the role of metal ions in biology, biochemistry, and medicine
Wu J,Chen W,Yin Y,Zheng Z,Zou G
Sun Jun 01 00:00:00 EDT 2014
Probing the cell death signaling pathway of HepG2 cell line induced by copper-1,10-phenanthroline complex.
Biometals : an international journal on the role of metal ions in biology, biochemistry, and medicine
Wu J,Chen W,Yin Y,Zheng Z,Zou G
Sun Jun 01 00:00:00 EDT 2014
Naringin inhibits TNF-α induced oxidative stress and inflammatory response in HUVECs via Nox4/NF-κ B and PI3K/Akt pathways.
Current pharmaceutical biotechnology
Li W,Wang C,Peng J,Liang J,Jin Y,Liu Q,Meng Q,Liu K,Sun H
Mon Aug 31 00:00:00 EDT 2015
Importin 7 and Nup358 promote nuclear import of the protein component of human telomerase.
PloS one
Frohnert C,Hutten S,Wälde S,Nath A,Kehlenbach RH
Fri Jan 02 00:00:00 EST 2015
Importin 7 and Nup358 promote nuclear import of the protein component of human telomerase.
PloS one
Frohnert C,Hutten S,Wälde S,Nath A,Kehlenbach RH
Fri Jan 02 00:00:00 EST 2015
Novel approaches for the identification of nuclear transport receptor substrates.
Methods in cell biology
Kimura M,Thakar K,Karaca S,Imamoto N,Kehlenbach RH
Thu Apr 16 00:00:00 EDT 2015
Chromatin-remodeling factors mediate the balance of sense-antisense transcription at the FGF2 locus.
Molecular endocrinology (Baltimore, Md.)
McEachern LA,Murphy PR
Tue Apr 01 00:00:00 EDT 2014
Chromatin-remodeling factors mediate the balance of sense-antisense transcription at the FGF2 locus.
Molecular endocrinology (Baltimore, Md.)
McEachern LA,Murphy PR
Tue Apr 01 00:00:00 EDT 2014
Pin1 promotes GR transactivation by enhancing recruitment to target genes.
Nucleic acids research
Poolman TM,Farrow SN,Matthews L,Loudon AS,Ray DW
Tue Oct 01 00:00:00 EDT 2013
Pin1 promotes GR transactivation by enhancing recruitment to target genes.
Nucleic acids research
Poolman TM,Farrow SN,Matthews L,Loudon AS,Ray DW
Tue Oct 01 00:00:00 EDT 2013
Resolution doubling in fluorescence microscopy with confocal spinning-disk image scanning microscopy.
Proceedings of the National Academy of Sciences of the United States of America
Schulz O,Pieper C,Clever M,Pfaff J,Ruhlandt A,Kehlenbach RH,Wouters FS,Großhans J,Bunt G,Enderlein J
Tue Dec 24 00:00:00 EST 2013
Resolution doubling in fluorescence microscopy with confocal spinning-disk image scanning microscopy.
Proceedings of the National Academy of Sciences of the United States of America
Schulz O,Pieper C,Clever M,Pfaff J,Ruhlandt A,Kehlenbach RH,Wouters FS,Großhans J,Bunt G,Enderlein J
Tue Dec 24 00:00:00 EST 2013
Several phenylalanine-glycine motives in the nucleoporin Nup214 are essential for binding of the nuclear export receptor CRM1.
The Journal of biological chemistry
Roloff S,Spillner C,Kehlenbach RH
Fri Feb 08 00:00:00 EST 2013
Several phenylalanine-glycine motives in the nucleoporin Nup214 are essential for binding of the nuclear export receptor CRM1.
The Journal of biological chemistry
Roloff S,Spillner C,Kehlenbach RH
Fri Feb 08 00:00:00 EST 2013
Identification of CRM1-dependent Nuclear Export Cargos Using Quantitative Mass Spectrometry.
Molecular & cellular proteomics : MCP
Thakar K,Karaca S,Port SA,Urlaub H,Kehlenbach RH
Fri Mar 01 00:00:00 EST 2013
Identification of CRM1-dependent Nuclear Export Cargos Using Quantitative Mass Spectrometry.
Molecular & cellular proteomics : MCP
Thakar K,Karaca S,Port SA,Urlaub H,Kehlenbach RH
Fri Mar 01 00:00:00 EST 2013
Long-distance intercellular connectivity between cardiomyocytes and cardiofibroblasts mediated by membrane nanotubes.
Cardiovascular research
He K,Shi X,Zhang X,Dang S,Ma X,Liu F,Xu M,Lv Z,Han D,Fang X,Zhang Y
Sat Oct 01 00:00:00 EDT 2011
Long-distance intercellular connectivity between cardiomyocytes and cardiofibroblasts mediated by membrane nanotubes.
Cardiovascular research
He K,Shi X,Zhang X,Dang S,Ma X,Liu F,Xu M,Lv Z,Han D,Fang X,Zhang Y
Sat Oct 01 00:00:00 EDT 2011
Intercellular transportation of quantum dots mediated by membrane nanotubes.
ACS nano
He K,Luo W,Zhang Y,Liu F,Liu D,Xu L,Qin L,Xiong C,Lu Z,Fang X,Zhang Y
Tue Jun 22 00:00:00 EDT 2010
Intercellular transportation of quantum dots mediated by membrane nanotubes.
ACS nano
He K,Luo W,Zhang Y,Liu F,Liu D,Xu L,Qin L,Xiong C,Lu Z,Fang X,Zhang Y
Tue Jun 22 00:00:00 EDT 2010
Regulation of FGF-2 by an endogenous antisense RNA: effects on cell adhesion and cell-cycle progression.
Molecular carcinogenesis
MacFarlane LA,Murphy PR
Wed Dec 01 00:00:00 EST 2010
Regulation of FGF-2 by an endogenous antisense RNA: effects on cell adhesion and cell-cycle progression.
Molecular carcinogenesis
MacFarlane LA,Murphy PR
Wed Dec 01 00:00:00 EST 2010
Regulation of fibroblast growth factor-2 by an endogenous antisense RNA and by argonaute-2.
Molecular endocrinology (Baltimore, Md.)
MacFarlane LA,Gu Y,Casson AG,Murphy PR
Thu Apr 01 00:00:00 EDT 2010
Regulation of fibroblast growth factor-2 by an endogenous antisense RNA and by argonaute-2.
Molecular endocrinology (Baltimore, Md.)
MacFarlane LA,Gu Y,Casson AG,Murphy PR
Thu Apr 01 00:00:00 EDT 2010
CHP2 activates the calcineurin/nuclear factor of activated T cells signaling pathway and enhances the oncogenic potential of HEK293 cells.
The Journal of biological chemistry
Li GD,Zhang X,Li R,Wang YD,Wang YL,Han KJ,Qian XP,Yang CG,Liu P,Wei Q,Chen WF,Zhang J,Zhang Y
Fri Nov 21 00:00:00 EST 2008
CHP2 activates the calcineurin/nuclear factor of activated T cells signaling pathway and enhances the oncogenic potential of HEK293 cells.
The Journal of biological chemistry
Li GD,Zhang X,Li R,Wang YD,Wang YL,Han KJ,Qian XP,Yang CG,Liu P,Wei Q,Chen WF,Zhang J,Zhang Y
Fri Nov 21 00:00:00 EST 2008