Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Store vial at 4° C prior to restoration. For extended storage aliquot contents and freeze at -20° C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4° C as an undiluted liquid. Dilute only prior to immediate use.
Beta Actin Antibody Peroxidase Conjugated antibody is directed against human beta Actin protein. A BLAST analysis was used to suggest that this antibody would react with beta Actin from a wide range of organisms, including most vertebrates and some yeast. Broad reactivity makes this antibody an excellent loading control.
Beta actin is one of six actin isoforms which have been identified. Beta actin is a non-muscle cytoskeletal protein in all human cell types and is involved in cell motility, structure, and integrity. There are six different actin isoforms in human. The beta isoform of actin, along with gamma actin, coexist in most cell types as components of the cyto-skeleton. Beta actins are cytoplasmic proteins ubuquitously expressed in all eukaryotic cells. Polymerization of globular actin (G-actin) leads to a structural filament (F-actin) in the form of two stranded helix. Actins are highly conserved proteins that are involved in cell motility, structure and integrity. Because beta actin is ubiquitously expressed in all eukaryotic cells, it is frequently used as a loading control for assays involving protein detection, such as Western blotting. Antibodies to beta-actin provide a specific and useful tool in studying the intracellular distribution of beta-actin and the static and dynamic aspects of the cytoskeleton.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: actin, beta; actin, beta, cytoplasmic; Actin, cytoplasmic 1; beta actin; beta cytoskeletal actin; Beta-actin; cytoplasmic beta-actin; cytoskeletal beta actin; cytoskeletal protein; PS1TP5-binding protein 1
Gene Aliases: ACTB; Actx; Beta-actin; BRWS1; E430023M04Rik; PS1TP5BP1
UniProt ID: (Sheep) P60713, (Bovine) P60712, (Human) Q96B34, (Pig) Q6QAQ1, (Guinea pig) Q71FK5, (Rat) P60711, (Mouse) P60710, (Chinese hamster) P48975
Entrez Gene ID: (Sheep) 443052, (Bovine) 280979, (Goat) 102179831, (Human) 60, (Pig) 414396, (Guinea pig) 100135470, (Rat) 81822, (Mouse) 11461, (Chinese hamster) 100689477
Molecular Function:
actin and actin related protein
actin or actin-binding cytoskeletal protein
cytoskeletal protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support