Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Reconstitute 40 µg lyophilized powder with 80 µl of deionized water (or equivalent) to antibody concentration of 0.5 mg/ml.
A number of mutations, identified in the gene encoding the β-amyloid precursor protein (βAPP), have been linked to early-onset Familial Alzheimers Disease. βAPP is cleaved sequentially by the proteolytic enzymes β-secretase and γ-secretase to produce β-amyloid (Aβ) peptides with the Aβ1-42(43) and the Aβ1-40 forms being the most prevalent. Secreted Aβ peptides can bind to scavenger receptors and the receptor for advanced glycation end products. Aβ peptides are degraded either via a reuptake mechanism followed by endosomal degradation or by an extracellular insulin-degrading enzyme. Extracellular accumulation of Aβ leads to formation of aggregates, fibrils, and eventually amyloid deposits called neuritic plaques, a hallmark of Alzheimer's disease. β-amyloid antibodies and peptides have been developed as tools for elucidating the biology of Alzheimers disease.GenScript Human β-Amyloid 1-42 Antibody (2C2G5), mAb, Mouse is produced from the hybridoma resulting from fusion of Sp2/0 myeloma and B-lymphocytes obtained from mouse immunized with a synthetic peptide corresponding to amino acids 36-42 of human β-Amyloid conjugated to KLH (Swiss Prot: P05067).
Beta amyloid is a 40-43 amino acid peptide cleaved from amyloid precursor protein by a protease, gamma-secretase. Beta Amyloid (1-40) (Amyloid beta 40 or Abeta 40) together with beta Amyloid (1-42) (Amyloid beta 42 or Abeta 42) are two major C-terminal variants of the amyloid beta protein, constituting the majority of amyloid beta proteins. Both the Amyloid beta 40 and Amyloid beta 42 alloforms are major components of the extracellular plaques found in Alzheimer's disease brain tissue.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: A beta 1 42; A β 1 42; AB42; ABPP; Alzheimer disease amyloid A4 protein homolog; Alzheimer disease amyloid protein; Amyloid; Amyloid b; Amyloid beta; amyloid beta (A4) precursor protein; Amyloid beta 1 42; Amyloid precursor protein; Amyloid β; Amyloid-beta (A4) precursor protein; Amyloid-beta A4 protein; Amyloid-beta precursor protein; APP; APPI; Aβ 1 42; B APP42; BAPP42; beta-amyloid peptide; beta-amyloid peptide(1-40); beta-amyloid peptide(1-42); beta-amyloid precursor protein; BetaAPP42; Cerebral vascular amyloid peptide; CVAP; peptidase nexin-II; PN-II; PreA4; Protease nexin-II; testicular tissue protein Li 2; β APP42; βAPP42
Gene Aliases: A4; AAA; ABETA; ABPP; AD1; APP; APPI; CTFgamma; CVAP; PN-II; PN2
UniProt ID: (Human) P05067
Entrez Gene ID: (Human) 351
Molecular Function: protease inhibitor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support