Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 5
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 µg/mL.
The protein encoded by this gene is a member of the EGF family of growth factors. It is synthesized primarily as a transmembrane precursor, which is then processed to mature molecule by proteolytic events. This protein is a ligand for the EGF receptor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: betacellulin; OTTHUMP00000160600; OTTHUMP00000219057; Probetacellulin
Gene Aliases: Bcn; Btc
UniProt ID: (Mouse) Q05928
Entrez Gene ID: (Mouse) 12223, (Rat) 64022
Molecular Function:
growth factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support