Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Positive Samples: 293T, A-549, Raji, Mouse lung, Mouse kidney, Rat spleen, Rat kidney
Immunogen sequence: LNRQLRGVSK EEVIRLKQKR RTLKNRGYAQ SCRFKRVQQR HVLESEKNQL LQQVDHLKQE ISRLVRERDA YKEKYEKLVS SGFRENGSSS DNPSSPEFFM
c-maf is the cellular counterpart of oncogenic v-maf that belongs to the family of basic region leucine zipper domain transcription factors. The leucine-zipper domain is involved in the interaction with LRPICD. There are two forms of human c-maf mRNA, c-maf-long and c-maf-short. It is identified in the genome of the acute transforming avian retrovirus AS42. c-maf targets are IL-4 in Th2 cells, the crystalline genes in lens fiber cells, insulin gene in islet, p53 and L7 where it exerts its transcriptional role through binding to a Maf recognition element (MARE). It regulates Th2 differentiation and lineage-specific hematopoiesis. c-maf is a transcription factor for IL-10 gene expression in LPS-activated macrophages. Chromosomal aberration involving maf is found in some forms of multiple myeloma. It is expressed in myeloma cell lines and resting monocytes/macrophages.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Avian musculoaponeurotic fibrosarcoma (MAF) protooncogene; C-Maf; c-Maf long form; c-maf proto-oncogene; MGC71685; Proto-oncogene c-Maf; T lymphocyte c-maf long form; Transcription factor Maf; Transcription factor Maf-2; v-maf avian musculoaponeurotic fibrosarcoma oncogene homolog; V-maf musculoaponeurotic fibrosarcoma oncogene homolog
Gene Aliases: 2810401A20Rik; A230108G15Rik; AW047063; AYGRP; c-MAF; CCA4; cMaf; CTRCT21; MAF; Maf2
UniProt ID: (Human) O75444, (Rat) P54844, (Mouse) P54843
Entrez Gene ID: (Human) 4094, (Rat) 54267, (Mouse) 17132
Molecular Function:
basic leucine zipper transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support