Search Thermo Fisher Scientific
GenScript
Reconstitute the lyophilized antibody with deionized water (or equivalent) to a final concentration of 0.5 mg/ml.
GenScript THE™ c-Myc Antibody [HRP], mAb, Mouse is immunoaffinity purified from mice ascites with protein A and then conjugated to horseradish peroxidase. Mouse Anti-c-Myc [HRP] Monoclonal Antibody is suitable for detecting the expression level of c-Myc in fusion proteins.
Myc is a proto-oncogene and encodes a nuclear phosphoprotein that plays a role in cell cycle progression, apoptosis and cellular transformation. The encoded protein forms a heterodimer with the related transcription factor MAX. This complex binds to the E box DNA consensus sequence and regulates the transcription of specific target genes. Amplification of this gene is frequently observed in numerous human cancers. Translocations involving this gene are associated with Burkitt lymphoma and multiple myeloma in human patients. There is evidence to show that translation initiates both from an upstream, in-frame non-AUG (CUG) and a downstream AUG start site, resulting in the production of two isoforms with distinct N-termini.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
Protein Aliases: 46-0603; c Myc; c-myc epitope tag; cmyc; Myc Tag; R950-25; R95025