Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAAGRLFLS RLRAPFSSMA KSPLEGVSSS RGLHAGRGPR RLSIEGNIAV GKSTFVKLLT KTYPEWHVAT EPVATWQNIQ AAGTQKACTA QSLGNLLDMM YREPARWSYT FQTFSFLSRL KVQLEPFPEK LLQARKPVQI FERSVYSDRY IFAKNLFENG SLSDIEWHIY QDWHSFLLWE FASRITLHGF IYLQASPQVC LKRLYQRARE EEKGIELAYL EQLHGQHEAW LIHKTTKLHF EALMNIPVLV LDVNDDFSEE VTKQEDLMRE VNTFVKNL (1-277 aa encoded by BC015757)
Mitochondrial deoxyguanosine kinase (DGUOK) is required for the phosphorylation of several deoxyribonucleosides and certain purine deoxykribonucleoside analogs widely employed as antiviral and chemotherapeutic agents. Purine deoxyribonucleoside analogs are extensively used in treatment of lymphoproliferative disorders. These compounds are administered as pro-drugs, and their efficiency is dependent on intracellular phosphorylation to the corresponding triphosphates. In mammalian cells, the phosphorylation of purine deoxyribonucleosides is mediated predominantly by 2 deoxyribonucleoside kinases: cytosolic deoxycytidine kinase (DCK) and mitochondrial deoxyguanosine kinase (DGUOK also known as DGK). DGUOK expression is ubiquitous, with highest levels in muscle, brain, liver and lymphoid tissues. Defects in DGUOK are a cause of mitochondrial DNA depletion syndrome (MDS). MDS is a clinically heterogeneous group of disorders characterized by a reduction in mitochondrial DNA (mtDNA) copy number. Primary mtDNA depletion is inherited as an autosomal recessive trait and may affect single organs, typically muscle or liver, or multiple tissues.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Deoxyadenosine kinase, mitochondrial; Deoxyguanosine kinase, mitochondrial
Gene Aliases: DGK; dGK1; dGK2; dGK3; DGUOK; MTDPS3
UniProt ID: (Human) Q16854, (Mouse) Q9QX60
Entrez Gene ID: (Human) 1716, (Mouse) 27369, (Rat) 297389
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support