Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This antibody reacts to cAMP (Cyclic Adenosine Monophosphate). No cross reactivity with other cyclic nucleotide or nucleoside phosphate.
Delta C2-alpha Tubulin has been identified as a biomarker for peripheral neuropathy. Delta C2-alpha tubulin is a C-terminally trimmed version of alpha-tubulin that lacks a carboxy-terminal glutamyl-tyrosine group.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alpha-tubulin 3; hum-a-tub1; hum-a-tub2; Tubulin alpha-1A chain; Tubulin alpha-3 chain; Tubulin B-alpha-1; tubulin, alpha 1a; tubulin, alpha, brain-specific
Gene Aliases: B-ALPHA-1; LIS3; TUBA1A; TUBA3
UniProt ID: (Human) Q71U36
Entrez Gene ID: (Human) 7846
Molecular Function:
tubulin
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support