Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
This gene encodes an essential eukaryotic translation initiation factor. The protein is required for the binding of the 43S complex (a 40S subunit, eIF2/GTP/Met-tRNAi and eIF3) to the 5' end of capped RNA.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: eIF-1A X isoform; eIF-4C; eukaryotic translation initiation factor 1A, X chromosome; Eukaryotic translation initiation factor 1A, X-chromosomal; eukaryotic translation initiation factor 1A, Y-linked; Eukaryotic translation initiation factor 4C; Putative eukaryotic translation initiation factor 1A
Gene Aliases: 1500010B24Rik; AI426898; eIF-1A; eIF-4C; EIF1A; EIF1AP1; EIF1AX; Eif1ay; EIF4C; RGD1560198
UniProt ID: (Human) P47813, (Mouse) Q8BMJ3
Entrez Gene ID: (Human) 1964, (Mouse) 66235, (Rat) 302697
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support