Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Immunogen sequence: MSYPADDYES EAAYDPYAYP SDYDMHTGDP KQDLAYERQY EQQTYQVIPE VIKNFIQYFH KTVSDLIDQK VYELQASRVS SDVIDQKVYE IQDIYENSWT KLTERFFKNT PWPEAEAIAP QVGNDAVFLI LYKELYYRHI YAKVSGGPSL EQRFESYYNY CNLFNYILNA DGPAPLELPN QWLWDIIDEF IYQFQSFSQY RCKTAKKSEE EIDFLRSNPK IWNVHSVLNV; Positive Samples: HepG2, HT-1080, NIH/3T3, Mouse heart, Mouse pancreas, Mouse liver; Cellular Location: Cytoplasm
Component of the eukaryotic translation initiation factor 3 (eIF-3) complex, which is required for several steps in the initiation of protein synthesis (PubMed:17581632, PubMed:25849773, PubMed:27462815). The eIF-3 complex associates with the 40S ribosome and facilitates the recruitment of eIF-1, eIF-1A, eIF-2:GTP:methionyl-tRNAi and eIF-5 to form the 43S pre-initiation complex (43S PIC). The eIF-3 complex stimulates mRNA recruitment to the 43S PIC and scanning of the mRNA for AUG recognition. The eIF-3 complex is also required for disassembly and recycling of post-termination ribosomal complexes and subsequently prevents premature joining of the 40S and 60S ribosomal subunits prior to initiation (PubMed:17581632). The eIF-3 complex specifically targets and initiates translation of a subset of mRNAs involved in cell proliferation, including cell cycling, differentiation and apoptosis, and uses different modes of RNA stem-loop binding to exert either translational activation or repression (PubMed:25849773). [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 66 kDa tyrosine-rich heat shock protein; 67 kDa polymerase-associated factor; eIEF associated protein HSPC021; EIF3L; Eukaryotic translation initiation factor 3 subunit 6-interacting protein; Eukaryotic translation initiation factor 3 subunit E-interacting protein; Eukaryotic translation initiation factor 3 subunit L; eukaryotic translation initiation factor 3, subunit 6 interacting protein; eukaryotic translation initiation factor 3, subunit L; HSP-66Y; PAF67
Gene Aliases: 0610011H21Rik; D15N1e; EIF3EIP; Eif3ip; EIF3L; EIF3S11; EIF3S6IP; HSP-66Y; HSPC021; HSPC025; MSTP005; PAF67
UniProt ID: (Human) Q9Y262, (Mouse) Q8QZY1
Entrez Gene ID: (Human) 51386, (Mouse) 223691, (Rat) 300069
Molecular Function:
translation factor
translation initiation factor
translational protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support