Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Immunogen sequence: QSVTSVAEKT KEQANAVSEA VVSSVNTVAT KTVEEAENIA VTSGVVRKED LRPSAPQQEG EASKEKEEVA EEAQSGGD; Positive Samples: SKOV3, HT-29; Cellular Location: Cytoplasm, centrosome, cytoskeleton, microtubule organizing center, perinuclear region, spindle
This gene encodes a member of the synuclein family of proteins which are believed to be involved in the pathogenesis of neurodegenerative diseases. Mutations in this gene have also been associated with breast tumor development.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Breast cancer-specific gene 1 protein; Gamma-synuclein; Persyn; SR; Synoretin; synuclein, gamma (breast cancer-specific protein 1)
Gene Aliases: BCSG1; C79089; PERSYN; PRSN; SNCG; SR
UniProt ID: (Human) O76070, (Mouse) Q9Z0F7
Entrez Gene ID: (Human) 6623, (Mouse) 20618
Molecular Function:
membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support