Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LVNQLSVDD VNVLTCATGT LSNLTCNNSK NKTLVTQNSG VEALIHAILR AGDKDDITEP AVCALRHLTS RHPEAEMAQN SVRLNYGIPA IVKLLNQPNQ WPLVKATIGL IRNLALCPAN HAPLQEAAVI PRLVQLLVKA HQDAQRHVAA GTQQPYTDGV RMEEIVEGCT GALHILARDP MNRMEIFRLN TIPLFVQLLY SSVENIQRVA AGVLCELAQD KEAADAIDAE GASAPLMELL HSRNEGTATY AAAVLFRISE DKNPDYRKRV SVELTNSLFK HDPAAWEAAQ SMIPINEPYG DDLDATYRPM YSSDVPLDPL EMHMDMDGDY PIDTYSDGLR PPYPTADHML A (396-745 aa encoded by BC011865)
JUP is a major cytoplasmic protein which is the only known constituent common to sub membranous plaques of both desmosomes and intermediate junctions. This protein forms distinct complexes with cadherins and desmosomal cadherins and is a member of the catenin family since it contains a distinct repeating amino acid motif called the armadillo repeat.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: catenin (cadherin-associated protein), gamma 80kDa; Catenin gamma; Desmoplakin III; Desmoplakin-3; gamma-catenin; gamma-catenin (plakoglobin); Junction plakoglobin; OTTHUMP00000164735
Gene Aliases: ARVD12; CTNNG; D930025P04Rik; DP3; DPIII; JUP; PDGB; PG; PKGB
UniProt ID: (Human) P14923, (Rat) Q6P0K8, (Mouse) Q02257
Entrez Gene ID: (Human) 3728, (Rat) 81679, (Mouse) 16480
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support