Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide Sequence: GYCPTMTRVL QGVLPALPQV VCNYRDVRFE SIRLPGCPRG VNPVVSYAVA LSCQCALCRR STTDCGGPKD HPLTCDDPRF QASSSSKAPP PSLPSPSRLP GPSDTPILPQ
This gene is a member of the glycoprotein hormone beta chain family and encodes the beta 7 subunit of chorionic gonadotropin (CG). Glycoprotein hormones are heterodimers consisting of a common alpha subunit and an unique beta subunit which confers biological specificity. CG is produced by the trophoblastic cells of the placenta and stimulates the ovaries to synthesize the steroids that are essential for the maintenance of pregnancy. The beta subunit of CG is encoded by 6 genes which are arranged in tandem and inverted pairs on chromosome 19q13.3 and contiguous with the luteinizing hormone beta subunit gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Choriogonadotropin subunit beta 7; chorionic gonadotropin beta 7 subunit; chorionic gonadotropin human; chorionic gonadotropin, beta polypeptide 7; human chorionic gonadotropin
Gene Aliases: CG-beta-a; CGB6; CGB7
UniProt ID: (Human) P0DN87
Entrez Gene ID: (Human) 94027
Molecular Function:
peptide hormone
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support