Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proposed to be involved in specific functions of the ESCRT machinery. Is required for efficient abscission during cytokinesis, but not for HIV-1 budding. The involvement in the MVB pathway is not established. Involved in recruiting VPS4A and/or VPS4B to the midbody of dividing cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Charged multivesicular body protein 8; CHMP8; hIST1; increased sodium tolerance 1 homolog; IST1 homolog; IST1, endosomal sorting complex required for transport-III component; MAPK activating protein PM28; MGC117220; overexpressed in lung cancer 1; pm28; Putative MAPK-activating protein PM28
Gene Aliases: 2400003C14Rik; AW536298; IST1; KIAA0174; mKIAA0174; OLC1; RGD1307799
UniProt ID: (Human) P53990, (Rat) Q568Z6, (Mouse) Q9CX00
Entrez Gene ID: (Human) 9798, (Rat) 307833, (Mouse) 71955
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support