Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Relative expression to ensure that the antibody binds to the antigen stated.
FIGURE: 1 / 6
Heat mediated antigen retrieval recommended prior to tissue staining.
This antibody is predicted to react with mouse, rat and macaque monkey based on sequence homology.
Store antibody at 4ºC for 1-2 weeks. For long-term storage, store at -20ºC.
HnRNP A1 belongs to the A/B subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins (hnRNPs). The hnRNPs are RNA binding proteins and they complex with heterogeneous nuclear RNA (hnRNA). These proteins are associated with pre mRNAs in the nucleus and appear to influence pre mRNA processing and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus, some seem to shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. hnRNP A1 has two repeats of quasi RRM domains that bind to RNAs. It is one of the most abundant core proteins of hnRNP complexes and it is localized to the nucleoplasm. This protein, along with other hnRNP proteins, is exported from the nucleus, probably bound to mRNA, and is immediately re imported.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Helix-destabilizing protein; Heterogeneous nuclear ribonucleoprotein A1; heterogeneous nuclear ribonucleoprotein A1B protein; heterogeneous nuclear ribonucleoprotein B2 protein; heterogeneous nuclear ribonucleoprotein core protein A1; hnRNP A1; hnRNP A1-like 3; hnRNP core protein A1; hnRNP core protein A1-like 3; HNRNP-I; HNRNPI; HNRPA1MGC102835; HNRPI; MGC102835; nuclear ribonucleoprotein particle A1 protein; pPTB; PTB-1; PTB-T; PTB2; PTB3; PTB4; putative heterogeneous nuclear ribonucleoprotein A1-like 3; RP11-78J21.1; single-strand DNA-binding protein UP1; Single-strand RNA-binding protein
Gene Aliases: ALS19; ALS20; hnRNP A1; hnRNP-A1; HNRNPA1; HNRPA1; HNRPA1L3; IBMPFD3; UP 1
UniProt ID: (Human) P09651
Entrez Gene ID: (Human) 3178
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support