Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody could both recognize unphosphorylated and phosphorylated iASPP.
Immunogen sequence: EGPVARPLS PTRLQPALPP EAQSVPELEE VARVLAEIPR PLKRRGSMEQ APAVALPPTH KKQYQQIISR LFHRHGGPGP GGPEPELSPI TEGSEARAGP PAPAPPAPIP PPAPSQSSPP EQPQSMEMRS VLRKAGSPRK ARRARLNPLV LLLDAALTGE LEVVQQAVKE MNDPSQPNEE GITALHNAIC GANYSIVDFL ITAGANVNSP DSHGWTPLHC AASCNDTVIC MALVQHGAAI FATTLSDGAT AFEKCDPYRE GYADCATYLA DVEQSMGLMN SGAVYALWDY SAEFGDELSF REGESVTVLR RDGPEETDWW WAALHGQEGY VPRNYFGLFP RVKPQRSKV (481-829 aa encoded by BC064913)
IASPP is one of the most evolutionarily conserved inhibitors of p53 (TP53), whereas ASPP1. and ASPP2. are activators of p53.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: inhibitor of apoptosis stimulating protein of p53; Inhibitor of ASPP protein; NFkB interacting protein 1; NFkB-interacting protein 1; PPP1R13B-like protein; ppp1r13l; Protein iASPP; protein phosphatase 1, regulatory (inhibitor) subunit 13 like; protein phosphatase 1, regulatory subunit 13 like; RelA-associated inhibitor; retinoic acid induced 4
Gene Aliases: AV260376; IASPP; NKIP1; PPP1R13BL; PPP1R13L; RAI; RAI4; wa3
UniProt ID: (Human) Q8WUF5, (Mouse) Q5I1X5
Entrez Gene ID: (Human) 10848, (Mouse) 333654, (Rat) 686781
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support