Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstituted by adding 1 mL sterile distilled water, spun down to remove insoluble particles, divided into small aliquots, frozen and stored at or below -20°C.
Homologous proteins of different species frequently share antigenic determinants. The degree of cross-reactivity is also dependent on the concentRations of the reactants and the sensitivity of the assay arrangement. This antiSerum has not been tested for cross-reactivity.
Diluted antiserum should be stored at 4°C, not refrozen, and preferably used the same day.
This gene encodes alpha-lactalbumin, a principal protein of milk. Alpha-lactalbumin forms the regulatory subunit of the lactose synthase (LS) heterodimer and beta 1,4-galactosyltransferase (beta4Gal-T1) forms the catalytic component. Together, these proteins enable LS to produce lactose by transfering galactose moieties to glucose. As a monomer, alpha-lactalbumin strongly binds calcium and zinc ions and may possess bactericidal or antitumor activity. A folding variant of alpha-lactalbumin, called HAMLET, likely induces apoptosis in tumor and immature cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: a-LACTA; alfaLA; Alpha-lactalbumin; lactalbumin; lactalbumin, alpha-; Lactose synthase B protein; lysozyme G; Lysozyme-like protein 7; MGC138521; MGC138523
Gene Aliases: LALBA; LYZG; LYZL7
UniProt ID: (Human) P00709
Entrez Gene ID: (Human) 3906
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support