Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
1 µg/mL of MA5-45553 was sufficient for detection of mGluR1/5 glutamate receptor in 20 µg of rat brain membrane lysate and assayed by colorimetric immunoblot analysis using goat anti-mouse IgG:HRP as the secondary antibody.|Detects approximately 130kDa (both mGluR1 and mGluR5).
This antibody was formerly sold as clone S75-33.
may play a role in synaptic transmission; may interact with mGluR1, another group I metabotropic glutamate receptor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: G protein coupled receptor family C group 1 member A; G protein coupled receptor, family C, group 1, member A; G protein coupled receptor, family C, group 1, member E; G protein-coupled receptor GRM5; G protein-coupled receptor, family C, group 1, member A; G protein-coupled receptor, family C, group 1, member E; glutamate receptor; glutamate receptor, metabotropic 1; glutamate receptor, metabotropic 5; metabotropic glutamate receptor (mGluR5); Metabotropic glutamate receptor 1; Metabotropic glutamate receptor 5; metabotropic glutamate receptor 5b; metabotropic glutamate receptor subtype 5b; metabotropic glutamate receptor type 5; mGluR1; mGluR5; mGlurR1a; protein phosphatase 1, regulatory subunit 85; protein phosphatase 1, regulatory subunit 86
Gene Aliases: 4930455H15Rik; 6430542K11Rik; AI850523; ENSMUSG00000075319; Glu5R; Gm10828; GPRC1A; GPRC1E; GRM1; GRM5; MGLU1; mGlu5; MGLUR1; MGLUR5; mGluR5b; nmf373; PPP1R85; PPP1R86; rcw; SCAR13; wobl
UniProt ID: (Human) Q13255, (Human) P41594, (Mouse) P97772, (Rat) P23385, (Rat) P31424, (Mouse) Q3UVX5
Entrez Gene ID: (Human) 2911, (Human) 2915, (Mouse) 14816, (Rat) 24414, (Rat) 24418, (Mouse) 108071
Molecular Function:
G-protein coupled receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support