Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSRVPAFLS AAEVEEHLRS SSLLIPPLET ALANFSSGPE GGVMQPVRTV VPVTKHRGYL GVMPAYSAAE DALTTKLVTF YEDRGITSVV PSHQATVLLF EPSNGTLLAV MDGNVITAKR TAAVSAIATK FLKPPSSEVL CILGAGVQAY SHYEIFTEQF SFKEVRIWNR TKENAEKFAD TVQGEVRVCS SVQEAVAGAD VIITVTLATE PILFGEWVKP GAHINAVGAS RPDWRELDDE LMKEAVLYVD SQEAALKESG DVLLSGAEIF AELGEVIKGV KPAHCEKTTV FKSLGMAVED TVAAKLIYDS WSSGK (1-314 aa encoded by BC018061)
Crystallins are separated into two classes: taxon-specific and ubiquitous. The former class is also called phylogenetically-restricted crystallins. The latter class constitutes the major proteins of vertebrate eye lens and maintains the transparency and refractive index of the lens. This gene encodes a taxon-specific crystallin protein that binds NADPH and has sequence similarity to bacterial ornithine cyclodeaminases. The encoded protein does not perform a structural role in lens tissue, and instead it binds thyroid hormone for possible regulatory or developmental roles. Multiple alternatively spliced transcript variants have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CDK108; crystallin, mu; Ketimine reductase mu-crystallin; mu-crystallin homolog; NADP-regulated thyroid-hormone binding protein; NADP-regulated thyroid-hormone-binding protein; thiomorpholine-carboxylate dehydrogenase
Gene Aliases: CRYM; DFNA40; THBP
UniProt ID: (Human) Q14894, (Rat) Q9QYU4, (Mouse) O54983
Entrez Gene ID: (Human) 1428, (Rat) 117024, (Mouse) 12971
Molecular Function:
oxidoreductase
reductase
metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support