Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Immunogen sequence: GYELLFQPE VVRIYISLLK ESKTPAILEA SAGAIQNLCA GRWTYGRYIR SALRQEKALS AIADLLTNEH ERVVKAASGA LRNLAVDARN KELIGKHAIP NLVKNLPGGQ QNSSWNFSED TVISILNTIN EVIAENLEAA KKLRETQGIE KLVLINKSGN RSEKEVRAAA LVLQTIWGYK ELRKPLEKEG WKKSDFQVNL NNASRSQSSH SYDDSTLPLI DRNQKSDKKP DREEIQMSNM GSNTKSLDNN YSTPNERGDH NKTLDRSGDL GDMEPLKGTT PLMQKI (546-830 aa encoded by BC010501)
Delta 1 Catenin is an efficient tyrosine kinase substrate implicated both in cell transformation by SRC and in ligand-induced receptor signaling through the EGF, PDGF, CSF-1 and ERBB2 receptors. The association of catenins to cadherins produces a complex which is linked to the actin filament network, and which seems to be of primary importance for cadherins cell-adhesion properties.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cadherin-associated Src substrate; Cadherin-associated Src substrate (CAS); CAS; Catenin (cadherin associated protein) delta 1; catenin (cadherin-associated protein), delta 1; Catenin delta-1; catenin src; p120 catenin; P120 CTN; P120 p120; P120 P120CTN; p120(cas); p120(ctn); p120-catenin
Gene Aliases: AA409437; AU019353; CAS; Catns; CTNND; CTNND1; KIAA0384; mKIAA0384; P120; p120(CAS); p120(CTN); P120CAS; P120CTN
UniProt ID: (Human) O60716, (Mouse) P30999
Entrez Gene ID: (Human) 1500, (Mouse) 12388, (Rat) 311163
Molecular Function: intermediate filament binding protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support