Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
In Immunoprecipitation this antibody co-precipitates cdk4/cdk6,.
p19 is a member of the INK4 family of cyclin-dependent kinase inhibitors. This protein has been shown to form a stable complex with CDK4 or CDK6, and prevent the activation of the CDK kinases, thus function as a cell growth regulator that controls cell cycle G1 progression. The abundance of the transcript of this gene was found to oscillate in a cell-cycle dependent manner with the lowest expression at mid G1 and a maximal expression during S phase. The negative regulation of the cell cycle involved in this protein was shown to participate in repressing neuronal proliferation, as well as spermatogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CDK inhibitor p19INK4d; cell cycle inhibitor, Nur77 associating protein; Cyclin-dependent kinase 4 inhibitor D; cyclin-dependent kinase 4 inhibitor D p19; cyclin-dependent kinase inhibitor 2D (p19, inhibits CDK4); inhibitor of cyclin-dependent kinase 4d; MTS1; p19-INK4d
Gene Aliases: CDKN2D; INK4D; p19; p19-INK4D
UniProt ID: (Human) P55273
Entrez Gene ID: (Human) 1032
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support