Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunoprecipitation: co-precipitates cdk4.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: p24 family protein gamma-3; p24gamma3; p27; Transmembrane emp24 domain-containing protein 7; transmembrane emp24 protein transport domain containing 7; UV36Sp6 ultraviolet B radiation-activated UV36
Gene Aliases: 3930401E15Rik; 5830493P14Rik; AU044611; AW545765; CGI-109; p24g3; p24gamma3; p27; TMED7; TRAM
UniProt ID: (Human) Q9Y3B3, (Rat) D3ZTX0
Entrez Gene ID: (Human) 51014, (Dog) 481436, (Mouse) 66676, (Rat) 252889
Molecular Function: vesicle coat protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support