Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSDASLRST STMERLVARG TFPVLVRTSA CRSLFGPVDH EELSRELQAR LAELNAEDQN RWDYDFQQDM PLRGPGRLQW TEVDSDSVPA FYRETVQVGR CRLLLAPRPV AVAVAVSPPL EPAAESLDGL EEAPEQLPSV PVPAPASTPP PVPVLAPAPA PAPAPVAAPV AAPVAVAVLA PAPAPAPAPA PAPAPVAAPA PAPAPAPAPA PAPAPAPDAA PQESAEQGAN QGQRGQEPLA DQLHSGISGR PAAGTAAASA NGAAIKKLSG PLISDFFAKR KRSAPEKSSG DVPAPCPSPS AAPGVGSVEQ TPRKRLR (1-316 aa encoded by BC067842)
This gene is part of a gene cluster on chromosome Xp11.23. The encoded protein contains a zinc finger motif often found in transcriptional regulators, however, its exact function is not known. Alternative splicing results in multiple transcript variants encoding the same protein.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Beckwith Wiedemann syndrome (WBS); Cyclin dependent kinase inhibitor 1C (CDKN1C); Cyclin-dependent kinase inhibitor 1C; cyclin-dependent kinase inhibitor 1C (p57, Kip2); Cyclin-dependent kinase inhibitor p57; IMAGE; p57 Kip2; p57Kip2; p57KIP2 A3sh
Gene Aliases: AL024410; BWCR; BWS; CDKI; CDKN1C; KIP2; p57; p57(kip2); p57KIP2; WBS
UniProt ID: (Human) P49918, (Mouse) P49919
Entrez Gene ID: (Human) 1028, (Mouse) 12577, (Rat) 246060
Molecular Function:
kinase inhibitor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support