Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
The recommended shelf life for this product is 1 year from date of receipt.
Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
Transcriptional repressor. Enhances MBD2-mediated repression. Efficient repression requires the presence of GATAD2B. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GATA zinc finger domain-containing protein 2A; Hp66alpha; MGC29315; MGC36975; p66a; Transcriptional repressor p66 alpha; Transcriptional repressor p66-alpha
Gene Aliases: 1110066C11Rik; BC031407; C80248; GATAD2A; p66alpha
UniProt ID: (Human) Q86YP4, (Mouse) Q8CHY6
Entrez Gene ID: (Human) 54815, (Mouse) 234366
Molecular Function:
DNA-binding transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support