Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The recommended shelf life for this product is 1 year from date of receipt.
Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
This gene encodes a zinc finger protein transcriptional repressor. The encoded protein is part of the methyl-CpG-binding protein-1 complex, which represses gene expression by deacetylating methylated nucleosomes. Mutations in this gene are linked to intellectual disability and dysmorphic features associated with cognitive disability.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GATA zinc finger domain-containing protein 2B; p66/p68; transcription repressor p66 beta component of the MeCP1 complex; transcription repressor p66 component of the MeCP1 complex; Transcriptional repressor p66-beta
Gene Aliases: AL118180; C430014D17Rik; GATAD2B; KIAA1150; mKIAA1150; MRD18; P66beta; p68
UniProt ID: (Human) Q8WXI9, (Mouse) Q8VHR5
Entrez Gene ID: (Human) 57459, (Mouse) 229542
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support