Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitute lyophilized product with 1 mL distilled water.
p97 ATPase/VCP (also described as valosin-containing protein) is implicated in homotypic membrane fusion events and is related to the N-ethyl maleimide-sensitive fusion protein NSF/Sec18p. Specificity with the p97 subunit of 15S Mg2+-ATPase. p97/VCP is involved in ubiquitin-proteasome dependent protein degradation processes and involvement was also found e.g. in the formation of neuronal inclusion bodies in neuro-degenerative diseseases (e.g. Parkinson's disease). Positive control: Immunoblots with protein extracts from all vertebrate tissues and cells.Reactivity on cultured cell lines: All vertebrate cells and tissues tested so far: e.g. Xenopus laevis; rabbit, rat, human; does not crossreact with the corresponding protein in budding yeast (Cdc48p).
CDK5RAP2 is a regulator of CDK5 (cyclin-dependent kinase 5) activity. The protein is localized to the centrosome and Golgi complex and interacts with CDK5R1 and pericentrin (PCNT). CDK5RAP2 plays a role in centriole engagement and microtubule nucleation, and has been linked to primary microcephaly and Alzheimer's disease. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: aging-associated protein 1; DAP-5; Death-associated protein 5; eIF-4-gamma 2; eIF-4G 2; eIF4G 2; EIF4G2; Eukaryotic translation initiation factor 4 gamma 2; eukaryotic translation initiation factor 4 gamma, 2; eukaryotic translation initiation factor 4G-like 1; FLJ41344; Novel APOBEC-1 target 1; p97; Translation repressor NAT1
Gene Aliases: AAG1; DAP5; EIF4G2; eif4g2.L; NAT1; OK/SW-cl.75; P97; XELAEV_18024574mg
UniProt ID: (Rabbit) P79398, (Human) P78344
Entrez Gene ID: (Rabbit) 100009236, (Human) 1982, (Rat) 361628, (Xenopus) 100127281
Molecular Function:
translation factor
translation initiation factor
translational protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support