Search Thermo Fisher Scientific
FIGURE: 1 / 2
Adding 1 mL of PBS buffer will yield a concentration of 100 µg/mL.
Cytokeratins are a subfamily of intermediate filament proteins and are characterized by a remarkable biochemical diversity, represented in human epithelial tissues by at least 20 different polypeptides. They range in molecular weight between 40 kDa and 68.
Protein Aliases: KRT1