Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Relative expression to ensure that the antibody binds to the antigen stated.
FIGURE: 1 / 8
Positive Control: mouse brain, rat brain
Predicted Reactivity: Human (93%), Xenopus laevis (84%), Pig (92%), Chicken (87%), Sheep (91%), Chimpanzee (93%), Bovine (91%)
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Synucleins are small, soluble proteins expressed primarily in neural tissue and in certain tumors. The family includes three known proteins: alpha-synuclein, beta-synuclein, and gamma-synuclein. The alpha- and beta-synuclein proteins are found primarily in brain tissue, where they are seen mainly in presynaptic terminals. The gamma-synuclein protein is found primarily in the peripheral nervous system and retina, but its expression in breast tumors makes it a candidate as a marker for tumor progression.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alpha-synuclein; Beta-synuclein; Gamma-synuclein; NACP; Non-A beta component of AD amyloid; non-A4 component of amyloid; Non-A4 component of amyloid precursor; Persyn; Phosphoneuroprotein 14; PNP 14; Sensory neuron synuclein; synuclein, gamma (breast cancer-specific protein 1)
Gene Aliases: AI838531; alphaSYN; betaSYN; C79089; NACP; Persyn; Snca; Sncb; Sncg; Syn
UniProt ID: (Rat) P37377, (Mouse) O55042, (Mouse) Q91ZZ3, (Rat) Q63754, (Rat) Q63544, (Mouse) Q9Z0F7
Entrez Gene ID: (Rat) 29219, (Mouse) 20617, (Mouse) 104069, (Rat) 113893, (Rat) 64347, (Mouse) 20618
Molecular Function:
membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support