Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitute in 100 µL of sterile water. Centrifuge to remove any insoluble material.
Specificity of this antibody: proEPO.
EPX mediates tyrosine nitration of secondary granule proteins in mature resting eosinophils. Shows significant inhibitory activity towards Mycobacterium tuberculosis H37Rv by inducing bacterial fragmentation and lysis. Allelic variant in EPX is associated with Japanese cedar pollinosis which is a type I allergic disease with ocular and nasal symptoms that develop paroxysmally on contact with Japanese cedar pollen. These symptoms, which occur seasonally each year, are typical features of allergic rhinitis, such as sneezing, excessive nasal secretion, nasal congestion, and conjunctival itching. Defects in EPX are the cause of eosinophil peroxidase deficiency (EPD). EPD is an autosomal recessive defect where anomalous eosinophils are characterized by nuclear hypersegmentation, hypogranulation, and negative peroxidase and phospholipid staining. Subcellular Location: Cytoplasmic granule.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Eosinophil peroxidase; EPO
Gene Aliases: EPER; EPO; EPP; EPX; EPX-PEN; EPXD
UniProt ID: (Human) P11678
Entrez Gene ID: (Human) 8288
Molecular Function:
peroxidase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support