Alomone Labs, LTD
FIGURE: 1 / 2
Reconstitution: 1 X 50 µL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).
MaxiK channels are large conductance, voltage and calcium-sensitive potassium channels which are fundamental to the control of smooth muscle tone and neuronal excitability. MaxiK channels can be formed by 2 subunits: the pore-forming alpha subunit and the modulatory beta subunit. The protein encoded by this gene is an auxiliary beta subunit which may partially inactivate or slightly decrease the activation time of MaxiK alpha subunit currents. Alternative splicing results in multiple transcript variants. A related pseudogene has been identified on chromosome 22.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BK beta 3; Calcium-activated potassium channel subfamily M subunit beta-3; Calcium-activated potassium channel subunit beta-3; kcnmb3 {ECO:0000312|EMBL:BAF79924.1}; Maxi K channel subunit beta-3; potassium large conductance calcium-activated channel, subfamily M beta member 3; potassium large conductance calcium-activated channel, subfamily M, beta member 4; slobeta3 (KCNMB3)
Gene Aliases: EG435726; Gm5707; Kcnmb3
UniProt ID: (Rat) A7VL23
Entrez Gene ID: (Mouse) 100502876, (Rat) 310303
Molecular Function:
voltage-gated ion channel
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support