Product References
The alcohol extracts of Sceptridium ternatum (Thunb.) Lyon exert anti-pulmonary fibrosis effect through targeting SETDB1/STAT3/p-STAT3 signaling.
Journal of ethnopharmacology
Zou X,Huang Z,Zhan Z,Yuan M,Zhang Y,Liu T,Hu X,Fan W,Chen P,Qin H,Zhang S,Xia Y,Zheng S,Pan Z,Huang P
Fri Sep 15 00:00:00 EDT 2023
Contribution of calcium dysregulation to impaired coronary artery contraction in Zucker diabetic fatty rats.
Clinical and experimental pharmacology & physiology
Liu L,Yang H,Kuang SJ,Zhang L,Zhou MY,Zeng P,Zhang MZ,Rao F,Zhou ZL,Deng CY
Wed Feb 01 00:00:00 EST 2023
Identification of necroptosis-related gene TRAF5 as potential target of diagnosing atherosclerosis and assessing its stability.
BMC medical genomics
Peng Z,Wang K,Wang S,Wu R,Yao C
Sat Jun 17 00:00:00 EDT 2023
LDHA mediated degradation of extracellular matrix is a potential target for the treatment of aortic dissection.
Pharmacological research
Wu X,Ye J,Cai W,Yang X,Zou Q,Lin J,Zheng H,Wang C,Chen L,Li Y
Tue Feb 01 00:00:00 EST 2022
Methamphetamine induces thoracic aortic aneurysm/dissection through C/EBPβ.
Biochimica et biophysica acta. Molecular basis of disease
Luo BY,Zhou J,Guo D,Yang Q,Tian Q,Cai DP,Zhou RM,Xu ZZ,Wang HJ,Chen SY,Xie WB
Thu Sep 01 00:00:00 EDT 2022
The Compatibility of Alisma and Atractylodes Affects the Biological Behaviours of VSMCs by Inhibiting the miR-128-5p/p21 Gene.
Evidence-based complementary and alternative medicine : eCAM
Wei W,Zhou YJ,Shen JL,Lu L,Lv XR,Lu TT,Xu PT,Xue XH
Tue Jul 19 00:00:00 EDT 2022
Platelet-Derived Microvesicles Promote VSMC Dedifferentiation After Intimal Injury via Src/Lamtor1/mTORC1 Signaling.
Frontiers in cell and developmental biology
Liu JT,Bao H,Fan YJ,Li ZT,Yao QP,Han Y,Zhang ML,Jiang ZL,Qi YX
Tue Oct 05 00:00:00 EDT 2021
Insulin Resistance Promotes the Formation of Aortic Dissection by Inducing the Phenotypic Switch of Vascular Smooth Muscle Cells.
Frontiers in cardiovascular medicine
Zheng H,Qiu Z,Chai T,He J,Zhang Y,Wang C,Ye J,Wu X,Li Y,Zhang L,Chen L
Sun May 01 00:00:00 EDT 2022
Insulin Resistance Promotes the Formation of Aortic Dissection by Inducing the Phenotypic Switch of Vascular Smooth Muscle Cells.
Frontiers in cardiovascular medicine
Zheng H,Qiu Z,Chai T,He J,Zhang Y,Wang C,Ye J,Wu X,Li Y,Zhang L,Chen L
Sun May 01 00:00:00 EDT 2022
Insulin Resistance Promotes the Formation of Aortic Dissection by Inducing the Phenotypic Switch of Vascular Smooth Muscle Cells.
Frontiers in cardiovascular medicine
Zheng H,Qiu Z,Chai T,He J,Zhang Y,Wang C,Ye J,Wu X,Li Y,Zhang L,Chen L
Sun May 01 00:00:00 EDT 2022
Insulin Resistance Promotes the Formation of Aortic Dissection by Inducing the Phenotypic Switch of Vascular Smooth Muscle Cells.
Frontiers in cardiovascular medicine
Zheng H,Qiu Z,Chai T,He J,Zhang Y,Wang C,Ye J,Wu X,Li Y,Zhang L,Chen L
Sun May 01 00:00:00 EDT 2022
The near-infrared dye IR-61 restores erectile function in a streptozotocin-induced diabetes model via mitochondrial protection.
Asian journal of andrology
Yue XF,Shen CX,Wang JW,Dai LY,Fang Q,Long L,Zhi Y,Li XR,Wang YW,Shen GF,Liu ZJ,Shi CM,Li WB
Fri Dec 03 00:00:00 EST 2021
The near-infrared dye IR-61 restores erectile function in a streptozotocin-induced diabetes model via mitochondrial protection.
Asian journal of andrology
Yue XF,Shen CX,Wang JW,Dai LY,Fang Q,Long L,Zhi Y,Li XR,Wang YW,Shen GF,Liu ZJ,Shi CM,Li WB
Fri Dec 03 00:00:00 EST 2021
Characterization of Carotid Smooth Muscle Cells during Phenotypic Transition.
Cells
Goikuria H,Freijo MDM,Vega Manrique R,Sastre M,Elizagaray E,Lorenzo A,Vandenbroeck K,Alloza I
Sun Mar 18 00:00:00 EDT 2018
Characterization of Carotid Smooth Muscle Cells during Phenotypic Transition.
Cells
Goikuria H,Freijo MDM,Vega Manrique R,Sastre M,Elizagaray E,Lorenzo A,Vandenbroeck K,Alloza I
Sun Mar 18 00:00:00 EDT 2018
Specific role of RhoC in tumor invasion and metastasis.
Oncotarget
Lang S,Busch H,Boerries M,Brummer T,Timme S,Lassmann S,Aktories K,Schmidt G
Fri Oct 20 00:00:00 EDT 2017
Specific role of RhoC in tumor invasion and metastasis.
Oncotarget
Lang S,Busch H,Boerries M,Brummer T,Timme S,Lassmann S,Aktories K,Schmidt G
Fri Oct 20 00:00:00 EDT 2017