Search Thermo Fisher Scientific
The Thermo Scientific Nexsa G2 X-Ray Photoelectron Spectrometer (XPS) System offers fully automated, high-throughput surface analysis, delivering the data to advance research and development or to solve production problems. The integration of XPS with ion scattering spectroscopy (ISS), UV photoelectron spectroscopy (UPS), reflected electron energy loss spectroscopy (REELS), and Raman spectroscopy, allows you to conduct true correlative analysis. The system now includes options for sample heating and sample biasing capabilities to increase the range of experiments now possible. The Nexsa G2 Surface Analysis System unlocks the potential for advances in materials science, microelectronics, nanotechnology development, and many other applications.
A new, low-power X-ray monochromator allows selection of the analysis area from 10 µm to 400 µm in 5 µm steps, ensuring data is collected from the feature of interest while maximizing the signal.
The high-efficiency electron lens, hemispherical analyzer, and detector allow for superb detectability and rapid data acquisition.
Bring sample features into focus with the Nexsa XPS System's patented optical viewing system and XPS SnapMap, which helps you pinpoint areas of interest quickly using a fully focused XPS image.
The patented dual-beam flood source couples low-energy ion beams with very low-energy electrons (less than 1 eV) to prevent sample charging during analysis, which eliminates the need for charge referencing, making the analysis of the data from insulating samples easy and reliable.
Go beyond the surface with a standard ion source or MAGCIS, the optional dual-mode monatomic and gas cluster ion source; automated source optimization and gas handling ensure excellent performance and experimental reproducibility.
Specialist sample holders for angle-resolved XPS, sample bias measurements, or for inert transfer from a glove box are available.
Instrument control, data processing, and reporting are all controlled from the Windows Software-based Avantage data system.
Fully software-controlled sample heating option, enabling temperature-dependent studies.
Animation: Thermo Scientific Nexsa surface analysis system
Analyzer type |
|
X-ray source type |
|
X-ray spot size |
|
Depth profiling |
|
Maximum Sample area |
|
Maximum sample thickness |
|
Vacuum system |
|
Optional accessories |
|
You will learn how the Nexsa G2 XPS System can be used to investigate a wide range of materials using XPS, supported by the additional analysis techniques on the system: UV photoelectron spectroscopy (UPS), ion scattering spectroscopy (ISS), reflected electron energy loss spectroscopy (REELS), Raman spectroscopy, and more.
You will learn how the Nexsa G2 XPS System can be used to investigate a wide range of materials using XPS, supported by the additional analysis techniques on the system: UV photoelectron spectroscopy (UPS), ion scattering spectroscopy (ISS), reflected electron energy loss spectroscopy (REELS), Raman spectroscopy, and more.
El desarrollo de baterías se realiza mediante análisis multiescala con microCT, SEM y TEM, espectroscopía Raman, XPS y visualización y análisis 3D digital. Aprenda cómo este enfoque proporciona la información estructural y química necesaria para crear mejores baterías.
La producción eficaz de metales requiere un control preciso de las inclusiones y precipitados. Nuestras herramientas automatizadas pueden realizar varias tareas cruciales para el análisis de metales, incluyendo el recuento de nanopartículas, el análisis químico EDS y la preparación de muestras de TEM.
La microestructura polimérica determina las características y el rendimiento del material a granel. La microscopía electrónica permite un análisis exhaustivo en microescala de la morfología y composición de los polímeros para aplicaciones de control de calidad e I+D.
Las ciencias geológicas están basadas en la observación uniforme y precisa de múltiples escalas de características dentro de las muestras de roca. SEM-EDS, combinado con software de automatización, permite el análisis directo a gran escala de la composición de la textura y los minerales para la investigación de la petrología y la mineralogía.
A medida que la demanda de petróleo y gas continúa, existe la necesidad constante de una extracción eficiente y eficaz de hidrocarburos. Thermo Fisher Scientific ofrece una amplia gama de soluciones de microscopía y espectroscopía para una gran variedad de aplicaciones de la ciencia del petróleo.
Los materiales tienen propiedades sustancialmente diferentes en la nanoescala y en la macroescala. Para estudiarlos, la instrumentación S/TEM se puede combinar con la espectroscopia de rayos X por dispersión de energía para obtener datos de resolución nanométrica, o incluso subnanométrica.
Los microrastros de las pruebas de las escenas del crimen se pueden analizar y comparar usando microscopía electrónica como parte de una investigación forense. Las muestras compatibles incluyen fragmentos de vidrio y pintura, marcas de herramientas, drogas, explosivos y residuos de armas de fuego.
Los catalizadores son cruciales para la mayoría de los procesos industriales modernos. Su eficacia depende de la composición microscópica y la morfología de las partículas catalíticas; EM con EDS es ideal para estudiar estas propiedades.
El diámetro, la morfología y la densidad de las fibras sintéticas son parámetros clave que determinan la vida útil y la funcionalidad de un filtro. La microscopía electrónica de barrido (SEM) es la técnica ideal para investigar rápida y fácilmente estas características.
La investigación de materiales novedosos presta cada vez más atención a la estructura de materiales de baja dimensión. La microscopía electrónica de transmisión de barrido con corrección de sonda y monocromación permite la adquisición de imágenes de materiales bidimensionales de alta resolución.
Todos los componentes de un vehículo moderno están diseñados para garantizar la máxima seguridad, eficacia y rendimiento. La caracterización detallada de materiales de automoción con microscopía electrónica y espectroscopía informa sobre decisiones cruciales sobre procesos, mejoras de productos y nuevos materiales.
La espectroscopía de fotoelectrones de rayos X (XPS) permite el análisis de superficie, proporcionando la composición elemental, así como el estado químico y electrónico de los 10 nm principales de un material. Con la realización de perfiles de profundidad, el análisis XPS se extiende a la visión de composición de las capas.
La espectroscopía de fotoelectrones de rayos X (XPS) permite el análisis de superficie, proporcionando la composición elemental, así como el estado químico y electrónico de los 10 nm principales de un material. Con la realización de perfiles de profundidad, el análisis XPS se extiende a la visión de composición de las capas.