Search Thermo Fisher Scientific
Discover the power of Applied Biosystems AgriSeq targeted genotyping by sequencing (GBS) solutions, designed to revolutionize the field of molecular breeding for agricultural applications. With our advanced genomic technologies, we offer a cost-effective and high-throughput approach to the analysis of genetic variation in plant and animal populations, helping breeders and researchers accelerate their programs and deliver improved species efficiently. Explore the possibilities and harness the potential of AgriSeq GBS for your agricultural genotyping needs.
The AgriSeq targeted GBS solution utilizes a highly efficient multiplexed PCR chemistry where hundreds to thousands of markers can be targeted and uniformly amplified in a single reaction. The amplicon libraries can then be barcoded and pooled for simultaneous sequencing of hundreds of samples on the Ion Torrent suite of next-generation sequencing (NGS) instruments. At a cost of pennies or less per genotype, AgriSeq targeted GBS is capable of generating up to 2.6 million genotypes per day from high-quality NGS data. Whether you’re looking to achieve more accurate marker-assisted breeding selection, or to improve your livestock parentage discrimination and trait monitoring, our solutions can help advance your plant and animal breeding programs.
AgriSeq genotyping is performed on the Ion Torrent NGS platform and is validated for 100–5,000 markers per sample. Using the Ion 540 chip, AgriSeq genotyping can produce up to 1.3 million unique genotype results per chip at an average read depth of 100x to achieve maximal genotype call rates. Combinations of different panels and species can also be processed on the same run.
Deliver high marker call rates with greater reproducibility across diverse sample sets to help ensure accurate selection. Marker call rates can exceed 96% with >99% inter- and& intra-run reproducibility, and AgriSeq targeted GBS data maintains a high concordance (>99%) when compared to the orthogonal genotyping technologies such as DNA microarrays.
Includes library generation, automated template prep, sequencing, and data analysis, which can be completed in ~2-3 days with <4 hours of hands-on time, enabling a single operator to process up to four 96- or 384-well plates per day. The Ion Chef System automates template preparation and chip loading, requiring only 15 min of hands-on time, followed by an overnight run.
AgriSeq high-density panels, ranging from 3K to 5K SNPs in size, are specifically designed to screen breeding populations for the most relevant SNPs with exceptional minor allele frequency and marker performance. Whether you need routine screening or other downstream genomic applications, these panels are a versatile solution.
Key features of AgriSeq ready-to-use panels include:
AgriSeq Panel Description |
Number of SNPs |
---|---|
Plant Panels |
|
USDA Wheat 5K |
5,000 |
Strawberry 4.5K |
4,500 |
Maize QA/QC |
160 |
Maize Trait |
259 |
Lettuce QA/QC/Trait |
215 |
Sunflower |
700 |
Chickpea |
5,000 |
Groundnut |
5,000 |
Pearl Millet |
4,000 |
Cotton |
4,500 |
Tomato 5K |
5,000 |
Pepper 5K |
5,000 |
Barley 3.5K |
3,500 |
Maize 3.6K |
3,600 |
Animal Panels |
|
ISAG Bovine Parentage |
554 |
ISAG Equine Parentage & Traits/Disorders |
950 |
Canine Traits & Disorders |
154 |
Canine Parentage & ID |
394 |
Feline Parentage & Traits/Disorders |
165 |
Caprine |
450 |
Ovine |
400 |
Pacific Oyster |
592 |
Coho Salmon |
434 |
Chinook Salmon |
599 |
Sockeye Salmon |
598 |
Eulachon (Candlefish) |
580 |
The AgriSeq Canine Traits and Disorders Panel contains 154 markers targeting clinically important genetic traits and disorders in canines. The panel includes 97 SNPs and complex markers such as 6 MNPs, 13 insertions, and 38 deletions. Most of these markers are common to all breeds, while some are breed specific for certain traits and conditions. The performance of the panel was verified using orthogonal testing, robustness testing, and field testing with diverse DNA samples from canine oral swabs. The panel enables multiplexed simultaneous genotyping of 154 markers to deliver reliable genotyping results for 136 genetic disorders and 18 traits using the AgriSeq targeted GBS workflow. The CanFam 3.1 reference genome is used for the analysis.
Panel Contains 379 SNPs and 2 deletion markers designed to deliver reliable genotyping results for parentage The AgriSeq Canine SNP Parentage and Identification Panel contains 379 SNPs and 2 deletion markers designed to deliver reliable genotyping results for parentage determination and identity verification of canines by targeted GBS. The panel enables multiplexed simultaneous genotyping using the AgriSeq targeted GBS workflow. The performance of the panel was verified using orthogonal testing, robustness testing, and field testing with diverse DNA samples from canine oral swabs. The panel was also qualified using the 2019 ISAG/ICAR 2nd SNP Typing Canine Comparison samples. The CanFam 3.1 reference genome is used for the analysis.
Figure 1. Performance of AgriSeq canine panels. Call rates were calculated for both panels after testing >180 diverse DNA field samples from canine oral swabs with the AgriSeq workflow. The mean call rate for the AgriSeq Canine Traits and Disorders Panel was 99.1% (A), and for the AgriSeq Canine SNP Parentage and Identification Panel was 99.2% (B), demonstrating the high performance obtained from customer samples.
The AgriSeq Feline Parentage and Identification Plus Traits and Disorders (PITD) Panel is a combined panel for determining parentage and identification (111 SNP markers) plus targeting 64 clinically important traits and disorders (43 SNP, 1 MNP, 4 insertion, and 16 deletion markers) in felines by targeted GBS. The performance of the panel was verified using orthogonal testing, robustness testing, and field testing with diverse DNA samples from feline oral swabs (Figure 4). The 111 SNP parentage and ID markers were also qualified using the 2019 ISAG/ICAR 2nd SNP Typing Feline Comparison samples. The comprehensive AgriSeq Feline PITD Panel is designed to deliver reliable genotyping results for parentage determination, including identity verification and prediction of 42 genetic disorders and 22 traits that are clinically important. When desired, additional custom markers referencing the Felis_catus_8.0 can be added if compatible.
The Applied Biosystems Bovine ISAG SNP Parentage Panel (2013) contains primers targeting the 200 ISAG-recommended SNPs available for simple multiplexed analysis using the AgriSeq targeted GBS workflow. The ISAG has standardized a set of 100 core SNPs and 100 additional SNPs with sufficient minor allele frequency (MAF) and genomic spacing for accurate bovine parentage determination across a wide range of breeds (Figure 5). You can download SNP information from the ISAG website [3]. Qualified using the 2015 ISAG/ICAR 3rd SNP Typing Bovine Comparison samples, our comprehensive Bovine ISAG Parentage Panel is designed to deliver reliable genotyping results for animal parentage testing. The Bos taurus reference genome is needed for analysis of the Bovine ISAG SNP Parentage Panel and can be obtained by contacting our technical support specialists.
Ion Torrent™ AgriSum Tool Kit (AST) plug-in aggregates data from the coverageAnalysis and variantCaller plugins for easy review of data. AST displays overall sequencing run summary metrics for easy review, enabling quality control or troubleshooting, if needed. Additionally, AST converts TVC data into genotype Martix, TOP, and TOP/BOTTOM formats for further analysis applications.
Agricultural genomics can be challenging for a number of reasons, including sequence complexity, high genetic diversity, and limited sequence availability. We offer end-to-end project management services through panel design, wet-lab validation, and data analysis to help ensure delivery of high-performance panels for your project. These low cost, proof of concept pilot projects are a great way to confirm the value of AgriSeq targeted GBS for your research.
IonCode Barcode Adapters are a family of products that provides 1536 unique barcode adapters designed for optimal performance with Ion Torrent semiconductor sequencers. When used in combination with the AgriSeq HTS Library, users can pool up to 1536 amplicon libraries and then conduct multiplexed sequencing analysis, which helps increase lab efficiency and reduce sequencing costs per sample.