Search Thermo Fisher Scientific
Oncomine Reporter produces clear and concise reports supporting cancer researchers and pathologists in the making of informed clinical cancer research decisions. The reports summarize relevant biomarker findings and link them with information from public sources about targeted therapies, guidelines, and global clinical trials.
The Oncomine Reporter workflow is streamlined into three easy steps, so that reports can be created in minutes.
Oncomine Reporter is based on global information aggregated in curated knowledgebase.
Additionally, the data are meticulously curated and updated monthly. After data are collected from various global data sources, a team of expert curation scientists manually reviews all candidate evidence. Two independent reviewers examine each piece of candidate evidence for context and standardization. The process has QC steps built-in at various stages. All approved evidence is then stored in a structured manner in a knowledgebase that serves to support reporting in Oncomine Reporter.
There are two options for obtaining Oncomine Reporter. The web-based Oncomine Reporter, available via Thermo Fisher Connect, enables a secure, scalable analysis platform with a pay-as-you-go reporting structure. Alternatively, the local server edition of Oncomine Reporter is available for those who are in a highly regulated environment or who run high quantities of reports. Learn more about the Local Server Edition from one of our knowledgeable staff members: