Visualize | Analyze | Manage | Understand | Innovate
Wherever imaging data sets need to be processed and managed, Thermo Scientific Software solutions offer abundant state-of-the-art image data processing, exploration and analysis features. Our adaptable software packages dedicated to life sciences, materials science, digital rock analysis, our service, and support options make complex visualization, analysis and data management of image data faster and easier for a wide range of customers.
Workflow using Avizo Software
Folha de estilo para cartões de instrumentos original
Visualization and analysis software
Software Amira
  • Importe e processe dados de imagem
  • Segmentação automática, separação de objetos, rotulagem
  • Analise e quantifique com estatísticas automatizadas
  • Crie imagens, animações e vídeos
Software Avizo
  • Compatível com vários dados/várias visualizações, vários canais, série temporal, dados muito grandes
  • Registro automático avançado em vários modos 2D/3D
  • Algoritmos de redução de artefatos
Software Pergeos
  • Compatível com vários dados/várias visualizações, vários canais, série temporal, dados muito grandes
  • Simulação de fluxo bifásico
  • Tomografia computadorizada de energia dupla (DECT)
Athena Software
Imaging Data Management
  • Ensure traceability of images, data, metadata and experimental workflows
  • Simplify your imaging workflow​
  • Improve collaboration
  • Secure and manage data access​
Amira-Avizo-python_1160x600
Explore the unlimited possibilities for processing your data
Amira-Avizo Software and PerGeos Software integrate Python 3.5. Many packages are available including NumPy and SciPy.
While providing a long list of packages, Python integration is not limited to those being installed by default.
Learn more
Amira-Avizo-machine-learning_1160x600
Accelerate image segmentation and improve image quality
Artificial intelligence (AI) methods, such as machine learning and deep learning, have proven to be powerful approaches for automating image segmentation and improving image quality.
Learn more
Amira-Avizo-recipes_1160x600
Increase your efficiency with the XTra library
The XTra library contains recipes, scripts, demos, and how-to videos dedicated to a wide range of specific use cases and workflows.
We are constantly adding XTras so you can find answers to all your questions.
Visit XTra Library
Services
Amira-Avizo-Introductory-training_1160x600
Introductory training
Shorten your learning curve and maximize your investment with this introductory training specifically designed for new users of Amira, Avizo and PerGeos Software.
The course consists of a lecture with hands-on sessions. The training material highlights the basic features and functionalities of Amira, Avizo and PerGeos Software.
Amira-Avizo-advanced-training_1160x600
Advanced training
Maximize your investment and reduce your time-to-results with this advanced training specifically designed for existing users of Amira, Avizo and PerGeos Software.
The course consists of a lecture with hands-on sessions. The training material highlights advanced features and functionalities of Amira, Avizo and PerGeos Software.
Amira-Avizo-Custom-Dev_1160x600
Custom development
With over 25 years of experience in 3D and image processing and hundreds of custom projects delivered to organizations small and large, Thermo Fisher Scientific can provide you with a solution tailored to fit your specific needs.
We can customize and expand our software solutions at various levels.