Search Thermo Fisher Scientific
Thermo Scientific Finntip Pipette Tips are a reliable and versatile solution for all your pipetting needs. Designed with precision and quality in mind, these tips offer accuracy and reproducibility in your experiments. With wide range of volumes available, from 0.2 µl to 10 mL, Finntip Pipette Tips cater to various sample volumes and pipette applications.
Tip type |
Benefits |
Recommended use |
---|---|---|
|
|
|
|
|
|
|
|
|
Micropoint Tip
|
|
|
|
|
|
|
|
|
Pipetting styles vary, so pair your Finnpipette with a compatible tip to achieve the system that performs best for your pipetting technique, with optimal precision and accuracy with every combination.
Requirement |
Finnpipette |
Suggested tip | Performance |
---|---|---|---|
Enhanced ergonomics | Finnpipette F1 |
Finntip Flex
|
|
Durability | Finnpipette F2
|
Finntip
|
|
High throughput | Finnpipette Novus |
Finntip Flex |
|
Finntip rack and Finnpipette are color coded by volume, making it easy to select compatible tips at a glance.
Color |
Volume Range |
|
---|---|---|
Pink | 10 µl |
|
Purple | 20 µl |
|
|
Turquoise | 50 µl |
Yellow | 200/250 µl |
|
|
Orange | 300 µl |
Blue | 1000 µl |
|
|
Green | 5 mL |
|
Red | 10 mL |
Package type |
Features |
---|---|
|
|
|
|
|
|
|
|
|
|
Individually wrapped |
|
For Laboratory Use. It is the customer’s responsibility to ensure that the performance of the product is suitable for customers’ specific uses or applications.