Search Thermo Fisher Scientific
Ion concentration measurement or ion-specific (ISE) measurements can be performed in every laboratory for a variety of sample types including water, food and beverage, pharmaceuticals, and biological samples.
To evaluate a sample's ion-specific performance, the best resource is slope S, also known from pH measurement. Whether you're looking to test ISE measurements through the direct method, dilution method, matrix match method, known addition method, or matrix factor method, our robust portfolio of ion-specific meters, electrodes, standards, and solutions are known for delivering accurate and reproducible ISE measurement results.
The ion selective electrode (ISE) is a useful and versatile sensor for testing ions in various types of samples, such as environmental, agricultural, industrial, biotechnical, pharmaceutical, food, wastewater, drinking water, and others. Typical ions of interest include ammonia, fluoride, nitrate, chloride, calcium, sodium, potassium, silver, sulfide, iodide, bromide, lead, cadmium, copper, and others.
|
|
|
|
To make an accurate measurement using an ISE, a suitable ISE method should be selected according to specific sample conditions.
Regulations vary between ion specific measurements, and it’s critical that you have instruments and electrodes that allow you to stay complaint with your industry standards. Below is a list of international standards for regulatory compliance requirements for a variety of industries.
International standards for regulatory compliance requirements:
Parameter | EPA | SM | ASTM | ISO | China MEP | USP | EP | AOAC | Other |
---|---|---|---|---|---|---|---|---|---|
pH | EPA 150.1 | 4500-H+(B) | D1293 (A,B) | 10523 | GB/T 6920 | <791> | 2.2.3 | 973.41 | FDA - 21 CFR 114.90 |
pH of Soil/Solid Waste | SW-846 9040C & 9045D | D4972 | 10390 | GB/T15555 | 994.16 - 994.18 | USDA NRCS | |||
Conductivity | EPA 120.1; SW-846 9050A | 2510 (B) | D1125 (A) | 7888 | n/a | <645> | 2.2.38 | 973.40 | PSS-78 (salinity) |
Dissolved Oxygen | 4500-O (G) | D888 (B, C) | 17289; 5814 | HJ506 | |||||
BOD | 5210 (B) | 5815 | HJ505 | 973.44 | |||||
Fluoride | SW-846 9214 | 4500-F (C ) | D1179 (B) | 10359 | GB7484 | ||||
Ammonia | 4500-NH3 (D, E) | D1426 (B) | 6778 | ||||||
Nitrate | Orion 601; SW-846 9210A | 4500-NO3 (D) | |||||||
Sulfide | SW-846 9215 | 4500-S2 (G) | D4658 | ||||||
Cyanide | SW-846 9213 | 4500-CN (F) | D2036 (A) | ||||||
Chloride | SW-846 9212 | 4500-Cl (D) | D512 (C) | GB11896 | |||||
Bromide | SW-846 9211 |
SM: Standard Methods for the Examination of Water and Wastewater. APHA, AWWA, WEF. | |
ASTM: American Society for the Testing and Materials | |
ISO: International Organization for Standardization | |
USP: United States Pharmacopeia | |
EP: European Pharmacopeia | |
China MEP: China Ministry of Environmental Protection | |
AOAC: Association of Official Analytical Chemists | |
NRCS: Natural Resources Conservation Service | |
PSS-78: Practical Salinity Scale of 1978 | |
FDA: U.S. Food and Drug Administration | |
USDA: United Stated Department of Agriculture |
Meet your sample analysis challenges with a combination of Thermo Scientific meters, electrodes, standards, and solutions, ideal for use with the following applications. Click on an icon below to learn more about these products.
The accuracy and reliability of your ISE measurements depend on the instrumentation you use. Learn more about how you can identify the best match based on your required features, performance, specifications, and budget
|
We offer electrodes, standards and electrode storage solutions, electrode cleaning solutions, and electrode filling solutions for your ISE measurement needs, Find te solutions that you need for your process. |
Validation and qualification (IQ/OQ) services for Thermo Scientific Orion bench and portable meters are currently available through Unity Lab Services in the United States ›