Search Thermo Fisher Scientific
The QuantStudio 1 Real-Time PCR System is a basic instrument for new and experienced users looking for a quality product at an affordable price. With a large interactive touchscreen and pre-optimized protocol templates, set up is easy.
Run reactions of up to 100 μL and detect differences as small as 1.5-fold in singleplex reactions, with results in approximately 40 minutes. The QuantStudio 1 system is also equipped with Connect, Thermo Fisher Scientific's cloud-based platform, allowing you to remotely monitor your runs in real time, analyze sophisticated data sets in minutes, securely store data, and share results online with colleagues across institutions and around the world.
In addition to the common features of all QuantStudio systems, the QuantStudio 1 system offers:
Three filters calibrated for FAM/SYBR Green/SYTO 9, VIC/HEX/TET/JOE, JUN/ROX/Texas Red dyes
Offers the three most commonly used excitation and emission filters: FAM, VIC, and ROX
Preoptimized protocol templates minimize new user training
Allows you to access, analyze, and share data anytime, anywhere
Category | Attribute | Description |
Format |
Fixed block |
|
Efficiency and productivity |
Small footprint |
|
Multiplexing |
3 |
|
Remote monitoring and Wi-Fi connectivity |
||
Cloud enabled |
||
Intuitive touchscreen |
||
Automation compatible |
||
Dimensions |
40 x 27 x 50 cm (H x W x D) |
|
Maximize uptime |
Smart Help |
|
Minimal calibration for easy maintenance |
||
Support |
Remote instrument diagnostics |
|
Access to remote service engineer |
For Research Use Only. Not for use in diagnostic procedures.