Search Thermo Fisher Scientific
Saccharomyces cerevisiae vacuolar membranes stained with the yeast vacuole membrane marker MDY-64. MDY-64 is available available separately (Cat. No. Y7536) or as part f our Yeast Vacuole Marker Sampler Kit (Cat. No. Y7531). The kit contains four additional fluorescent vacuolar lumen stains.