Search Thermo Fisher Scientific
DNA extracts from camptothecin-treated HL-60 cells were separated on an agarose gel and stained with SYBR® Green I Nucleic Acid Gel Stain (Cat. No. S7563, S7567, S7585). The 200 to 5,000 bp DNA fragments characteristic of apoptotic cells (which appear as "ladders") are clearly visualized with this sensitive nucleic acid stain. Cell preparations were gifts of Zbigniew Darzynkiewicz, Cancer Research Institute, New York Medical College.
1% Agarose gel containing 16S and 23S ribosomal RNA (rRNA). SYBR® Green II RNA gel stain. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›
Imaging autophagy in live HeLa cells with CellLight® reagents for mitochondria and lysosomes: Go ›
Traditional fluorescence imaging of oxidative stress and apoptosis using CellROX® Deep Red Reagent (Cat. No. C10422) and CellEvent™ Caspase-3/7 (Cat. No. C10423) Go ›