Search Thermo Fisher Scientific
FluoCells prepared slide #6 (F35925) showing a fixed, permeabilized, and labeled muntjac skin fibroblast. Mitochondria were labeled with mouse anti–OxPhos Complex V inhibitor protein antibody and visualized using orange-fluorescent Alexa Fluor® 555 goat anti–mouse IgG antibody (Cat. no. A21422). F-actin was labeled with green-fluorescent Alexa Fluor® 488 phalloidin (Cat. no. A12379), and the nucleus was stained with TO-PRO®-3 iodide (Cat. no. T3605, pseudocolored magenta).
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›
Endothelial cells. Go ›
Mouse Anti-Alpha Tubulin Monoclonal Antibody (Cat. No. A11126) Go ›
Muntjac fibroblast labeled with probes for actin and the nucleus. Go ›
A prometaphase muntjac skin fibroblast stained with Alexa Fluor® 350 phalloidin, an anti–a-tubulin antibody and an anti–cdc6 peptide antibody. Go ›
Simultaneous detection of expression of five genes in a whole mount Drosophila embryo by fluorescence in situ hybridization (FISH) with five RNA probes Go ›
1% Agarose gel containing 16S and 23S ribosomal RNA (rRNA). SYBR® Green II RNA gel stain. Go ›
Multicolor fluorescence analysis of muntjac fibroblasts. Go ›